Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 900

Ph Industrial Electrodes   to   Thermo Scientific Laboratory Refrigerator

Image
  Product Detail
More Detail

Ph Industrial Electrodes

Buy Ph Industrial Electrodes Items Including:
  • Flat pH surface electrode, for use in trace HF (2% concentration)
  • In-Line pH Electrodes with CPVC Housing (YO-27106-00)
  • In-Line pH Electrodes with CPVC Housing, 100Ω RTD (YO-27106-10)
  • In-Line pH Electrodes with CPVC Housing, 1 kΩ RTD (YO-27106-20).

15 Position Stirrers

Buy 15 Position Stirrers Items Including:
  • Thermo Scientific Cimarec i Telesystem 15 position Stirrer with Telemo (50088034)
  • Thermo Scientific Cimarec i Telesystem 15 position Stirrer with Telemo (50088036)
  • IKA RO 15 Position Stirrer, 100-240V
  • Multi-position Stirrer, 15 position 100-240V.

Resistivity Transmitter

Buy Resistivity Transmitter Items Including:
  • Cole-Parmer 300 Conductivity/Resistivity/Temperature 1/8-DIN Transmitt
  • Cole-Parmer 350 Conductivity/Resistivity/Temperature 1/8- DIN Controll
  • +GF+ Signet Single-Channel Conductivity/Resisitivity Transmitter, Fiel
  • H Comm Module
  • Angle adj Adapter.

Aquapro Electrodes

Buy Aquapro Electrodes Items Including:
  • Thermo Scientific Orion AquaPro Professional pH electrode, glass body, (9104APWP)
  • Thermo Scientific Orion AquaPro Professional pH electrode, glass body, (9102AP)
  • Thermo Scientific Orion AquaPro Professional pH electrode, glass body, (9103APWP).

Turbine Stirrer

Buy Turbine Stirrer Items Including:
  • Turbine Stirrer (YO-50703-35)
  • Turbine Stirrer (YO-50703-36).

Inline Flow Switch

Buy Inline Flow Switch Items Including:
  • Stainless steel liquid flow switch for high inline pressures; 0.5 GPM (YO-32774-42)
  • Stainless steel liquid flow switch for high inline pressures; 1.0 GPM (YO-32774-44)
  • UL-approved Liquid Flow Switch for High Inline Pressures; 0.10 GPM (YO-32778-06).

Gp High Pressure Pumps

Buy Gp High Pressure Pumps Items Including:
  • Pump, Cole-Parmer Large Peristaltic Metering Pump, Max flow of 28.5 GP (YO-74203-41)
  • Pump, Cole-Parmer Large Peristaltic Metering Pump, Max flow of 17.4 GP (YO-74203-42).

Labs Testing Equipment

Buy Labs Testing Equipment Items Including:
  • Test Tube Rack 8 mm Diameter X 86 Holes
  • Test Tube Rack 10 mm Diameter X 86 Holes
  • Test Tube Rack 12 mm Diameter X 58 Holes
  • Test Tube Rack 14 mm Diameter X 58 Holes
  • Test Tube Rack 16 mm Diameter X 32 Holes

Combination Ph Electrodes

Buy Combination Ph Electrodes Items Including:
  • Replacement combination pH electrode, gel filled with built-in tempera (103636)
  • Accumet pH Electrode, Combination, Microprobe, Refillable, Glass Body,
  • Accumet pH Electrode, Combination, General Purpose, Long-neck, Gel-fil.

Emergency Eyewash and Shower Station Mats

Buy Emergency Eyewash and Shower Station Mats Items Including:
  • Wearwell Eyewash Station Mat, 30" x 36", Green/Yellow
  • Wearwell Eyewash Station Mat, 30" x 48", Green/Yellow
  • Bradley Combination Showers-Shower/eye/facewash station
  • Guardian Equipment Freeze-Resistant Safety Station with eyewash.

Digital Shaking Water Baths

Buy Digital Shaking Water Baths Items Including:
  • Lab Companion 17L Reciprocal Shaking Water Bath - 120VAC, 60Hz (YO-12054-00)
  • Lab Companion 17L Reciprocal Shaking Water Bath - 230VAC, 50Hz (YO-12054-05)
  • Lab Companion 25L Reciprocal Shaking Water Bath - 120VAC, 60Hz (YO-12054-10).

Bulk Packed Platinum Cured Silicone Tubing

Buy Bulk Packed Platinum Cured Silicone Tubing Items Including:
  • Platinum-Cured Silicone Tubing, 1/50"ID x 1/12"OD, 25 ft/pack
  • Platinum-Cured Silicone Tubing, 1/32"ID x 3/32"OD, 25 ft/pack
  • Platinum-Cured Silicone Tubing, 1/2"ID x 11/16"OD, 25 ft/pack.

Helmet Type with Face Protection

Buy Helmet Type with Face Protection Items Including:
  • Pro-Shield™ Face Shield Kit with helmet, safety glasses, protective.

Nalgene Graduates

Buy Nalgene Graduates Items Including:
  • Thermo Scientific Nalgene graduated polymethylpentene cylinder, 100 mL
  • Thermo Scientific Nalgene graduated polymethylpentene cylinder, 250 mL
  • Thermo Scientific Nalgene graduated polymethylpentene cylinder, 500 mL.

Wire Recorders

Buy Wire Recorders Items Including:
  • DataNet PT100 sensor (2 wires) - PTFE cable length 2.5 meter (12753)
  • DataNet PT100 sensor (2 wires) - PTFE cable length 4 meter (12752).

In Process Ph Meter

Buy In Process Ph Meter Items Including:
  • T-Fitting for In-Line Process pH and ORP Electrodes, Kynar (YO-27003-86)
  • Etatron HDS pH Pump Control System, 5 L/hr, 10 bar max pressure, wall-
  • Etatron DLX pH/ORP Pump Control System, 1 L/hr, 15 bar max pressure, w (DLX-pH-RX/MBB) (YO-56022-40).

Accuform Warning Biohazard (Symbol) Signs

Buy Accuform Warning Biohazard (Symbol) Signs Items Including:
  • Safety Sign, Warning - Biohazard, 7" X 10", Adhesive Vinyl
  • Safety Sign, Warning - Biohazard, 7" X 10", Aluminum
  • Safety Sign, Warning - Biohazard, 10" X 14", Adhesive Vinyl.

Compact Chiller Benchtop

Buy Compact Chiller Benchtop Items Including:
  • Polyscience compact benchtop chiller, -20 to 40°C, 680 watts at 0 (YO-12930-10)
  • Polyscience compact benchtop chiller, -20 to 40°C, 680 watts at 0 (YO-12930-15)
  • Polyscience low temp benchtop chiller, -20 to 40°C, 750 watts at 0.

Chemical Equipment

Buy Chemical Equipment Items Including:
  • Fixed-angle 24 x 1.5/2.0-mL Rotor with chemical resistant (PTFE) coati
  • Thermo Scientific Nalgene Chemically resistant sealing cap assembly; 3
  • Replacement EtO dose chemical indicator strips 25/bx (AN87).

Heidolph Brinkmann Tuttnauer Sterilizers

Buy Heidolph Brinkmann Tuttnauer Sterilizers Items Including:
  • Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Analog 0.7 cu ft, 12 (23210100)
  • Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Analog, 0.7 cu ft, 2 (23210151)
  • Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Analog 0.8 cu ft, 12 (23210304).

2 Pvc Globe Valve

Buy 2 Pvc Globe Valve Items Including:
  • Bronze globe valve; 2" NPT(F) (GLV0700780)
  • Hayward electrically Actuated Ball Valve, 3-way, 2" PVC (HCTN1200STV/EAU-28)
  • Wide-Range PVC Relief Valve, 1/2" NPT(F) (RVD050V-PV)
  • PVC Check Valve, 1/2" NPT(F) connections (CKM050V-PV).

Cole-Parmer Automated Self-Cleaning Vacuum Systems

Buy Cole-Parmer Automated Self-Cleaning Vacuum Systems Items Including:
  • Automated self-cleaning dry vacuum system, 115 VAC.

Helmets Face Protection

Buy Helmets Face Protection Items Including:
  • Pro-Shield™ Face Shield Kit with helmet, safety glasses, protective
  • Pro-Shield™ Face Shield with helmet
  • Weight-Balancing Arc Flash Face Shield and Helmet Kit, with safety gla
  • Optional Attachable Task Light with Clip for Arc Flash Face Shield.

Single-Use Luer

Buy Single-Use Luer Items Including:
  • PendoTECH PressureMAT Single-Use Sensor, Luer Fitting, Polycarbonate (PRESS-S-000)
  • Single Use Temperature Sensor with Luer Fitting
  • PendoTECH PressureMAT 1/4" x 1/4" Hose Barb to Luer Fitting, Polycarbo (PDKT-103-03).

Advantec Litmus Papers

Buy Advantec Litmus Papers Items Including:
  • Litmus Blue Test Paper, 10 Booklets of 20 Strips.

44 Gauge Thermocouple Wire

Buy 44 Gauge Thermocouple Wire Items Including:
  • Type T 16-gauge thermocouple wire; PVC/PVC outer/inner insulation; 100 (T16-5-502 | 08541-34)
  • Thermocouple Wire ,TYPE-J ,30-GAUGE, FEP Insulation,100-ft Spool (J30-1-506)
  • Thermocouple Wire ,TYPE-J ,30-GAUGE, FEP Insulation,1000-ft Spool (J30-1-506).

Laboratory Refractometers

Buy Laboratory Refractometers Items Including:
  • Digital Refractometer, Brix, 45.0 to 95.0% (300033)
  • Digital Refractometer, Brix, 0.0 to 95.0% (300034)
  • Digital Refractometer, Brix, 0.0 to 60.0%, 0.0 to 28.0% Salinity, 1.33 (300035).

96 Well Plate Covers

Buy 96 Well Plate Covers Items Including:
  • 96-well Deep Well Plate, Polypropylene, 48/pack (YO-07904-02)
  • 96-well Deep Well Plate, Polypropylene, 24/pack (YO-07904-04)
  • 96-well Deep Well Plate, Polystyrene, 32/pack (YO-07904-06)
  • Costar 96-Well Standard Microplates, Nontreated, PP, Flat Well.

Fluid Stirrer

Buy Fluid Stirrer Items Including:
  • Flexible shaft for overhead stirrers (YO-54400-45)
  • Remote controller for Heidolph electronic overhead mixers (YO-54400-55)
  • VISCO JET stainless steel impeller, 120mm (YO-54400-70)
  • Lightnin® 316 SS mixing shaft, 5/8" dia x 36"L (208559316).

Thermo Scientific Laboratory Refrigerator

Buy Thermo Scientific Laboratory Refrigerator Items Including:
  • Laboratory undercounter refrigerator (single door); 5.4 CU
  • FT; 115V (REL-404A)
  • Laboratory space-saver refrigerator (single door); 11.5 CU FT; 115V (REL-1204A-20)
  • Laboratory space-saver refrigerator (single door); 11.5 CU FT; 200/230 (REL-1204D).



Ph Industrial Electrodes to Thermo Scientific Laboratory Refrigerator

>