Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 10

Cole-Parmer Barbed Reducing Y Connectors   to   500 Ml Volumetric Flask

Image
  Product Detail
More Detail

Cole-Parmer Barbed Reducing Y Connectors

Buy Cole-Parmer Barbed Reducing Y Connectors Items Including:
  • Barbed reducer tee, PVDF, 1/8x1/4", 10/pack (YO-30705-08)
  • Barbed Fittings; Reducing T Connector, 316 SS, 1/4" x 1/8" Tubing ID (YO-31208-45)
  • Barbed Fittings; Reducing T Connector, 316 SS, 1/4" x 3/8" Tubing ID (YO-31208-46).

Pipe Thread to Barb Fitting

Buy Pipe Thread to Barb Fitting Items Including:
  • Barbed fittings, NPT male pipe adapter, Black PP, 1/8" OD x 3/16" NPT, (YO-06444-16)
  • Barbed fittings, NPT male pipe adapter, Black PP, 1/4" OD x 3/16" NPT, (YO-06444-18).

Plastic Sample Vials

Buy Plastic Sample Vials Items Including:
  • Thermo Scientific Nalgene Sample vials with snap caps, 12 mL
  • Snap-cap sample vials, 11.1 mL (19271519-144)
  • Snap-cap sample vials, 18.5 mL (19272519-144)
  • Snap-cap sample vials, 25.9 mL (19273619-144)
  • Snap-cap sample vials, 37.0 mL (19275019-144).

Male Luer Lock Metal Fitting

Buy Male Luer Lock Metal Fitting Items Including:
  • Luer fittings, 316 SS fittings, male luer lock x 1/8" hose barb (YO-31507-25)
  • Luer fittings, 316 SS fittings, male luer lock x 1/16" hose barb (YO-31507-26)
  • Nickel-plated brass fittings, male luer lock x 1/16" hose barb (YO-31507-62).

Parker Distributors Ball Valves

Buy Parker Distributors Ball Valves Items Including:
  • Parker two-way straight ball valve, stainless steel, 1/8" compression
  • Parker two-way right angled ball valve, stainless steel, 1/4" compress
  • Parker two-way right angled ball valve, stainless steel, 3/8" compress.

Rotary Piston Vacuum Pumps

Buy Rotary Piston Vacuum Pumps Items Including:
  • Vacuubrandreg Rotary Vane Vacuum Pump, 1.6 cfm, 1.5 X 10-3/sup (698123)
  • Vacuubrandreg Rotary Vane Vacuum Pump, 1.6 cfm, 1.5 X 10-3/sup (698120)
  • Vacuubrandreg Rotary Vane Vacuum Pump, 4.0 cfm, 1.5 X 10-3/sup (698133).

Oakton Ph Buffer Solution Msds

Buy Oakton Ph Buffer Solution Msds Items Including:
  • Oakton pH buffer pouches; pH 4.01, 20/pk
  • Oakton pH buffer pouches; pH 7.00, 20/pk
  • Oakton pH buffer pouches; pH 10.00, 20/pk
  • Oakton 4.01 pH buffer, 500-mL
  • Oakton 1.68 pH buffer, 500-mL

Oakton Ion Selective Electrodes

Buy Oakton Ion Selective Electrodes Items Including:
  • Ion Selective Electrode (YO-35802-00)
  • Ion Selective Electrode (YO-35802-02)
  • SOLUTION KIT AMMONIUM (YO-35802-52)
  • SOLUTION KIT AMMONIUM (YO-35802-53)
  • SOLUTION KIT CHLORIDE (YO-35802-62)

Myron L Conductivity/Resistivity/Tds Meters and Testers

Buy Myron L Conductivity/Resistivity/Tds Meters and Testers Items Including:
  • MYRON L 512T5 analog TDS meter
  • MYRON L 532T1 analog TDS meter
  • MYRON L EP analog conductivity/resistivity meter
  • MYRON L TechPro II™ TP1 conductivity/TDS meter
  • MYRON L 512M5 analog conductivity meter

Cole-Parmer Flow-through Ph Cells and Electrodes

Buy Cole-Parmer Flow-through Ph Cells and Electrodes Items Including:
  • Cole-Parmer Flow Cell for Ultrapure Water, 316 SS
  • pH electrode, combination, double-junction, sealed, for flow-through c (YO-05662-44)
  • pH flow-through cell, 6000 µL, 7 mm ID, glass (YO-05662-40)
  • Electrode cable, detachable, 3-ft, BNC (YO-05662-55).

8 X10 Glass Fiber Filter Paper

Buy 8 X10 Glass Fiber Filter Paper Items Including:
  • Advantec Grade GA55 Glass Fiber Filters, 125mm dia; 100/box
  • Advantec Grade GB140 Glass Fiber Filters; 47mm dia; 100/box
  • Advantec Grade GB140 Glass Fiber Filters; 90mm dia; 100/box
  • Advantec Grade GB140 Glass Fiber Filters; 125mm dia; 100/box.

Flexible Liner Pumps

Buy Flexible Liner Pumps Items Including:
  • Polypropylene flexible liner pump, Nordel rubber liner, 1 GPM, 115/230
  • Polypropylene flexible liner pump, Nordel rubber liner, 3 GPM, 115/230
  • Polypropylene flexible liner pump, Nordel rubber liner, 5 GPM, 115/230.

Cole-Parmer 100% Nylon Knit Glove Liners

Buy Cole-Parmer 100% Nylon Knit Glove Liners Items Including:
  • Glove liners, full-fingered, small, 12 pairs/pk
  • Glove liners, full-fingered, medium, 12 pairs/pk
  • Glove liners, full-fingered, large, 12 pairs/pk
  • Glove liners, full-fingered, x-large, 12 pairs/pk
  • Glove liners, half-fingered, medium, 12 pairs/pk.

Staining Jars and Dishes for Histology

Buy Staining Jars and Dishes for Histology Items Including:
  • Glass Coplin Staining Jar with glass cover (YO-48585-30)
  • Coplin Staining Jar (YO-48585-40)
  • Columbia Staining Jar
  • Columbia Staining Jar, case of 6 (YO-48585-52)
  • Dish and Cover for 48585-00 (YO-48585-01)

Sterile Sampling Bags

Buy Sterile Sampling Bags Items Including:
  • Whirl-Pak sterile sampling bag, 18 oz
  • Whirl-Pak sterile sampling bag, 24 oz
  • Whirl-pak Sterile Sampling Bag, 69 oz 500/bx
  • Whirl-Pak sterile sampling bag with white labeling area, 2 oz
  • Whirl-Pak sterile sampling bag with white labeling area, 18 oz.

Hose Barb X Female Npt Adapter

Buy Hose Barb X Female Npt Adapter Items Including:
  • Hose Barb Adapter, PTFE, 1/8" NPT(F) X 1/8" ID (F-FH0101)
  • Hose Barb Adapter, PTFE, 1/8" NPT(F) X 1/4" ID (F-FH0102)
  • Hose Barb Adapter, PTFE, 1/4" NPT(F) X 1/8" ID (F-FH0201).

Npt to 1 4 Barb Fitting

Buy Npt to 1 4 Barb Fitting Items Including:
  • Barbed fittings, NPT male pipe adapter, Natural PP, 1/4" NPT x 1/4" I (YO-30610-38)
  • Barbed fittings, NPT male branch tees, Natural PP, 1/8" NPT x 1/4" ID, (YO-30610-72).

Wheaton Scintillation Vials

Buy Wheaton Scintillation Vials Items Including:
  • 20-mL Glass Scintillation Vials; size 22 urea cap/polyetheylene disc l (986548)
  • 20-mL Glass Scintillation Vials; size 24 urea cap/polyethylene disc li (986568)
  • 20-mL Polyethylene Scintillation Vials; urea cap/metal foil liner, cap (986702).

Check Valve 1 4 Flare

Buy Check Valve 1 4 Flare Items Including:
  • Poppet Check Valve, 1/4" NPT(F) connections (CV-1-1144)
  • PP Miniature Check Valve, 1/4" hose barb, Viton diaphragm
  • PVDF Miniature Check Valve, 1/4" hose barb, Viton diaphragm
  • PVC Check Valve, 1/4" NPT(F) connections (CKD025V-PV).

Micro Stir Bars

Buy Micro Stir Bars Items Including:
  • Micro Stir Bar, 6.35mm L x 3mm dia (YO-08545-02)
  • Scienceware Spinbar Stirring Bar, PTFE, Flea Micro, Yellow, 6.35x3mm
  • Scienceware Spinbar Stirring Bar, PTFE, Flea Micro, Blue, 6.35x3mm
  • Scienceware Spinbar Stirring Bar, PTFE, Flea Micro, Red, 7x2mm.

Pyrex Brand Fiber Glass Wool

Buy Pyrex Brand Fiber Glass Wool Items Including:
  • Fiber Glass Wool, 12-lb pack (3950).

Stainless Steel Barb Fittings

Buy Stainless Steel Barb Fittings Items Including:
  • Compression To Thread Adapter, Stainless Steel, 6mm OD Tubing X 1/4" N (YO-31528-24)
  • Compression Straight Union, Stainless Steel, 10mm OD Tubing (YO-31528-35)
  • Compression Reducing Union, Stainless Steel, 12mm X 6mm OD Tubing (YO-31528-41).

Plastic Ball Check Valve

Buy Plastic Ball Check Valve Items Including:
  • Hayward True Union Ball Check Valve, 1/2" socket and NPT(F) connectors
  • Hayward True Union Ball Check Valve, 3/4" socket and NPT(F) connectors
  • Hayward True Union Ball Check Valve, 1-1/2" socket and NPT(F) connecto.

Oakton Ph 11 Meter

Buy Oakton Ph 11 Meter Items Including:
  • Oakton pH 11 meter with "All-in-One" electrode (35811-71)
  • Oakton pH 11 meter only
  • Oakton pH 11 and DJ electrode with NIST-traceable Calibration
  • Oakton pH 700 Benchtop Meter, double-junction refillable glass pH elec (pH 700) (YO-35419-11).

Uvex Safety Glasses

Buy Uvex Safety Glasses Items Including:
  • Uvex Protege™ Safety Glasses, Black frame, Clear lens, Ultra-dura c
  • Uvex Slate Safety Glasses, Gunmetal Frame, Hardcoat Lens Coating, Clea
  • Uvex Astro OTG Safety Glasses, Clear Lens, Black Frame
  • Uvex Astro OTG Safety Glasses, Clear Lens, Blue Frame.

10 Ml Volumetric Flask

Buy 10 Ml Volumetric Flask Items Including:
  • PYREX Brand 5640 Volumetric Flask; 500 mL, pack of 6
  • PYREX Brand 5580 Volumetric Flask; 500 mL, pack of 6
  • PYREX Brand 5642 Volumetric Flask; 1000 mL, pack of 1
  • Thermo Scientific Nalgene Class B polypropylene volumetric flask, 50 m.

Pyrex Brand 9826 Culture Tubes

Buy Pyrex Brand 9826 Culture Tubes Items Including:
  • Pyrex Brand 9826 culture tube; 9 mL, pack of 144
  • Pyrex Brand 9826 culture tube; 11 mL, pack of 144
  • Pyrex Brand 9826 culture tube; 15 mL, pack of 144
  • Pyrex Brand 9826 culture tube; 25 mL, pack of 96
  • Pyrex Brand 9826 culture tube; 34 mL, pack of 96.

Nalgene Test Tube Racks Plastic

Buy Nalgene Test Tube Racks Plastic Items Including:
  • Thermo Scientific Nalgene Unwire™ test tube rack for 13-mm tubes, w
  • Thermo Scientific Nalgene Unwire™ test tube rack for 13-mm tubes, o
  • Thermo Scientific Nalgene Unwire™ test tube rack for 13-mm tubes, y.

A&D Gf Toploading Balances

Buy A&D Gf Toploading Balances Items Including:
  • A&D GF Toploading Balance, 410g x 0.001g External Calibration, RS232,
  • A&D GF Toploading Balance, 610g x 0.001g External Calibration, RS232,
  • A&D GF Toploading Balance, 810g x 0.001g External Calibration, RS232,.

500 Ml Volumetric Flask

Buy 500 Ml Volumetric Flask Items Including:
  • PYREX Brand 5640 Volumetric Flask; 500 mL, case of 12
  • PYREX Brand 5580 Volumetric Flask; 500 mL, case of 12
  • PYREX Brand 5660 Lifetime Red Volumetric Flask; 500 mL, case of 12
  • Thermo Scientific Nalgene Class B polypropylene volumetric flask, 500.



Cole-Parmer Barbed Reducing Y Connectors to 500 Ml Volumetric Flask

>