Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 100
Ph Standard Solutions
to
Ph Meter 440 Manual
Image
Product Detail
More Detail
Ph Standard Solutions
Buy Ph Standard Solutions Items Including:
Standardizing solution: 0.1 M NaNO3 (920706)
Standardizing solution: 0.1 M CaCl2 (922006)
Standardizing solution: NaF as F, 100 ppm (940907)
Standardizing solution: NaCl as Na, 100 ppm (941107)
Standardizing solution: NaCl as Cl, 100 ppm (941707).
More Details
Pharmafluor Reducing Compression Elbows
Buy Pharmafluor Reducing Compression Elbows Items Including:
Pharmafluor Reducing Compression Elbow, PFA, 1/8 X 1/4 (YO-31001-16)
Pharmafluor Reducing Compression Elbow, PFA, 1/4 X 3/8 (YO-31001-17).
More Details
50 Ml Beaker
Buy 50 Ml Beaker Items Including:
Pyrex Brand 1000 Griffin Low-Form Beaker, 50 mL, 12/pk (1000-50 425160)
Pyrex Brand 1000 Griffin Low-Form Beaker, 50 mL, 48/cs (1000-50)
Thermo Scientific Nalgene Griffin low-form beaker of PFA, 50 mL
Kimax blue-coded Griffin beakers; 50 mL, 12/cs.
More Details
Ysi Conductivity Standards
Buy Ysi Conductivity Standards Items Including:
YSI conductivity calibration standard, 10,000 µS
YSI conductivity calibration standard, 100,000 µS.
More Details
Parker Hannifin Threaded Plugs
Buy Parker Hannifin Threaded Plugs Items Including:
Threaded plug, PFA, 1/8" male NPT (YO-31321-13)
Threaded plug, PFA, 1/4" male NPT (YO-31321-14)
Threaded plug, PFA, 3/8" male NPT (YO-31321-15)
Threaded plug, PFA, 1/2" male NPT (YO-31321-16).
More Details
Thermo Scientific Nalgene Heavy-Duty Polypropylene Bottles
Buy Thermo Scientific Nalgene Heavy-Duty Polypropylene Bottles Items Including:
Thermo Scientific Nalgene heavy-duty polypropylene bottle, 4 L
Thermo Scientific Nalgene heavy-duty polypropylene bottle, 250 mL (DS2126-0250)
Thermo Scientific Nalgene Replacement polypropylene cap, 38-430.
More Details
Humboldt Saybolt Viscometer Bath
Buy Humboldt Saybolt Viscometer Bath Items Including:
Viscometer Tube, Stainless steel, for Saybolt Viscometer Bath (K22309)
Socket wrench for Saybolt Viscometer Bath (K22050)
Viscometer Tube, Brass, for Saybolt Viscometer Bath (K22009)
Receiving flask, Pyrex glass, 60 mL, for Saybolt Viscometer Bath.
More Details
Saybolt Viscometer Baths
Buy Saybolt Viscometer Baths Items Including:
Viscometer Tube, Stainless steel, for Saybolt Viscometer Bath (K22309)
Socket wrench for Saybolt Viscometer Bath (K22050)
Viscometer Tube, Brass, for Saybolt Viscometer Bath (K22009)
Receiving flask, Pyrex glass, 60 mL, for Saybolt Viscometer Bath.
More Details
Lamotte Spectroscopy and Spectrophotometers
Buy Lamotte Spectroscopy and Spectrophotometers Items Including:
Aluminum test kit (3641-SC)
Ammonia nitrogen test kit (365901-SC)
Chlorine test kit (6739-G)
Fluoride test kit (3647-01-SC)
Iron test kit (3668-SC)
More Details
Viscosity Cups
Buy Viscosity Cups Items Including:
Standard Ford Dip Viscosity Cup, orifice #3
Standard Ford Dip Viscosity Cup, orifice #4
EZ series viscosity cup #3 with certificate
EZ series viscosity cup #4
EZ series viscosity cup #4 with certificate
More Details
Corning Hot Plate
Buy Corning Hot Plate Items Including:
Corning Digital Hot Plate, 10" x 10", 120 VAC
Corning Digital Stirring Hotplate, 5" x 7", 120 VAC
Corning Digital Stirring Hotplate, 5" x 7", 220 VAC
Corning Digital Stirring Hotplate, 10" x 10", 120 VAC.
More Details
Ph Meter for Small Sample Volumes
Buy Ph Meter for Small Sample Volumes Items Including:
Thermo Scientific Orion Star LogR pH Meter Kit with PerpHect glass-bod
Thermo Scientific Orion Star LogR pH Meter with PerpHect glass-body pH
Thermo Scientific Orion Star LogR pH Meter with PerpHect ROSS glass-bo
Hach waterproof ISFET pH meter kit.
More Details
Cole-Parmer Asme-Bpe Equal Cross Clamp Fittings
Buy Cole-Parmer Asme-Bpe Equal Cross Clamp Fittings Items Including:
316L Stainless Steel Asme-bpe Sanitary Clamp Equal Cross, 20ra Mechani (YO-30606-33)
316L Stainless Steel Asme-bpe Sanitary Clamp Equal Cross, 20ra Mechani (YO-30606-34).
More Details
Cole-Parmer Wildco Aquatic Kick Nets
Buy Cole-Parmer Wildco Aquatic Kick Nets Items Including:
Wildco Kick Net Multifilment 800-900 um (425-K13).
More Details
Fisher Scientific Nalgene Trays
Buy Fisher Scientific Nalgene Trays Items Including:
Thermo Scientific Nalgene polypropylene autoclaving tray, 1 L
Thermo Scientific Nalgene polypropylene autoclaving tray, 2 L
Thermo Scientific Nalgene polypropylene autoclaving tray, 3 L
Thermo Scientific Nalgene polypropylene autoclaving tray, 5 L.
More Details
Pipette Tips
Buy Pipette Tips Items Including:
Thermo Scientific Finntip 200 Ext Pipette Tips, 5 to 200 µL, 400/bo
Thermo Scientific Finntip ® 300 Pipette Tips, 5-300 µL, 10x96/rack (9401-250)
Thermo Scientific Finntip 10 mL Sterile Pipette Tips, 2 to 10 mL, 5x24.
More Details
Metric Ptfe Tubing
Buy Metric Ptfe Tubing Items Including:
PTFE tubing, 6.00 x 7 mm, 25 ft/pk (YO-06363-10)
PTFE tubing, 8.00 x 9 mm, 25 ft/pk (YO-06363-20)
PTFE tubing, 10.0 x 11 mm, 25 ft/pk (YO-06363-30)
PTFE Tubing, 1.48 x 3mm, 25 ft/pk (YO-06605-52)
PTFE Tubing, 7.98 x 10mm, 25 ft/pk (YO-06605-56).
More Details
Ph Meter for Small Samples
Buy Ph Meter for Small Samples Items Including:
Sterile Sample Bottles, 100 Pk
Thermo Scientific Orion Star LogR pH Meter Only
Thermo Scientific Orion Star LogR pH Meter Kit with PerpHect glass-bod
WTW pH 1970i Heavy-Duty pH Meter
Oakton pH meter with probe
More Details
High-Efficiency Polypropylene Absolute-Rated Filter Bags
Buy High-Efficiency Polypropylene Absolute-Rated Filter Bags Items Including:
Polypropylene absolute-rated high-efficiency filter bags; 18-1/2"L, 25 (255028-03)
Polypropylene absolute-rated high-efficiency filter bags; 18-1/2"L, 50 (255029-03).
More Details
Barnstead Nanopure Water System
Buy Barnstead Nanopure Water System Items Including:
Nanopure Life Science Systems (RO/Distilled Feed) Consumables Bundle (D502144)
Organic-Free RO or Distilled Feed Cartridge Pack for Thermo Scientific (D50280)
Thermo Scientific Barnstead Pretreatment Cartridge for reverse osmosis (D50250).
More Details
I/P Main Body/Occlusion Bed Assemblies
Buy I/P Main Body/Occlusion Bed Assemblies Items Including:
Main body/occlusion bed assemblies with rear bearing, PSF/CRS housing
Main body/occlusion bed assemblies with rear bearing, PSF/SS housing m.
More Details
Heating Mantles
Buy Heating Mantles Items Including:
Electrothermal Heating Mantle for 50-mL flask, PTFE coated, 9.25"W x 4 (CMU0050/CEX1)
Electrothermal Heating Mantle for 500-mL flask, PTFE coated, 11"W x 5
(CMU0500/CEX1).
More Details
Cleaning Solutions
Buy Cleaning Solutions Items Including:
General-purpose ultrasonic cleaning solution, 0.9 L (1 qt) bottle
Cole-Parmer Micro-90 cleaning solution, 5 gallon bottle
Cole-Parmer Micro-90 cleaning solution, 55 gallon drum
Cole-Parmer Micro-90 cleaning solution, case of twelve 1 liter bottles.
More Details
Accessories for Extech Do700 Multiparameter Meter
Buy Accessories for Extech Do700 Multiparameter Meter Items Including:
Replacement DO electrode
Replacement Membrane Caps, 3/PK
Replacement pH electrode.
More Details
Ohaus Balance Printer Cables
Buy Ohaus Balance Printer Cables Items Including:
Print Cable for use with Ohaus SF40A Printer (On older Voyger or Explo
OHAUS RS-232 CABLE AND ADAPTER FOR COMPACT THERMAL PRINTER 11901-47 (80252580)
Ohaus RS-232 Cable And Adapter For Compact Dot Matrix Printer 11901-59 (Accessory) (YO-11901-63).
More Details
250 Ml Volumetric Flask
Buy 250 Ml Volumetric Flask Items Including:
Corning 250 mL PC Flask w/vented cap, 50/CS
PYREX Brand 5660 Lifetime Red Volumetric Flask; 250 mL, pack of 6
PYREX Brand 5660 Lifetime Red Volumetric Flask; 250 mL, case of 12
Thermo Scientific Nalgene Class B polypropylene volumetric flask, 250.
More Details
Dynalon Solvent Venting Wash Bottles
Buy Dynalon Solvent Venting Wash Bottles Items Including:
Azlon solvent venting wash bottle for isopropanol, 250 mL
Azlon solvent venting wash bottle assortment pack, 250 mL
Azlon solvent venting wash bottle for isopropanol, 500 mL
Azlon solvent venting wash bottle with write-on area, 250 mL.
More Details
Wheaton Headspace Vials
Buy Wheaton Headspace Vials Items Including:
10 mL Headspace Vial with 20 mm Aluminum Seal Convenience Pack, PTFE/S (W225278-01)
10 mL Headspace Vial with 20 mm Aluminum Seal Convenience Pack, PTFE/G (W225278-02)
Pressure Release Aluminum Seals, 20 mm, PTFE/Silicone Septa, 100/CS (W224215).
More Details
Manual Beckman Ph Meter 350
Buy Manual Beckman Ph Meter 350 Items Including:
Thermo Scientific Orion 3-star benchtop pH meter kit, 115/220 VAC
Thermo Scientific Orion 2-star benchtop pH meter, 115/220V
Thermo Scientific Orion 2-star benchtop pH meter, value kit 115/220V
Thermo Scientific Orion 3-Star handheld pH meter kit.
More Details
Ph Meter 440 Manual
Buy Ph Meter 440 Manual Items Including:
WTW pH 1970i Heavy-Duty pH Meter
Thermo Scientific Orion 3-star benchtop pH meter, 115/220 VAC
Thermo Scientific Orion 3-star benchtop pH meter kit, 115/220 VAC
Thermo Scientific Orion 2-star benchtop pH meter, 115/220V
YSI Ecosense pH Meter Only.
More Details
70618
results
<<<Previous
96
97
98
99
100
101
102
103
104
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
20
30
40
50
60
70
80
85
86
87
88
89
90
91
92
93
94
95
96
97
98
99
100
101
102
103
104
105
106
107
108
109
110
111
112
113
114
120
130
140
150
160
170
180
190
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Ph Standard Solutions to Ph Meter 440 Manual
>
31125329-s-toc_2 31125329stoc2