Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 11
25 Ml Volumetric Flask
to
Welding Fume Hoods
Image
Product Detail
More Detail
25 Ml Volumetric Flask
Buy 25 Ml Volumetric Flask Items Including:
PYREX Brand 5640 Volumetric Flask; 25 mL, case of 12
PYREX Brand 5580 Volumetric Flask; 25 mL, case of 12
PYREX Brand 5660 Lifetime Red Volumetric Flask; 250 mL, case of 12
PYREX Brand 5660 Lifetime Red Volumetric Flask; 25 mL, case of 12.
More Details
Peristaltic Pump
Buy Peristaltic Pump Items Including:
Pump, Adustable Peristaltic Pump, 4 GPD (0.6 LPH), 100 PSI (6.8 BAR), (XP004LANX)
Pump, Adustable Peristaltic Pump, 15 GPD (2.4 LPH), 100 PSI (6.8 BAR), (XP015LANX)
Pump, Adustable Peristaltic Pump, 23 GPD (3.6 LPH), 100 PSI (6.8 BAR), (XP023LANX).
More Details
Plastic Hand-Operated Heavy-Duty Acid-Transfer Hand Pumps
Buy Plastic Hand-Operated Heavy-Duty Acid-Transfer Hand Pumps Items Including:
Hand-operated acid-transfer pump, 6 strokes/gallon, glass carboy adapt (236PF3-G)
Hand-operated acid-transfer pump, 6 strokes/gallon, 2" IPS adapter (236PF3S)
Hand-operated acid-transfer pump, 6 strokes/gallon, male buttress adap (236PF3B).
More Details
Wind Speed Meters
Buy Wind Speed Meters Items Including:
Kestrel 2000 Pocket Wind Meter; Measures Wind Speed Temperature Wind
Kestrel 1000 Pocket Wind Meter; Measures Wind Speed Maxwind Gust And.
More Details
Electric Drum Pump
Buy Electric Drum Pump Items Including:
Cole-Parmer Drum Pump Air Motor, 22 cfm at 92 psi
Cole-Parmer Drum Pump Tube Set, PP, 39"
Cole-Parmer Drum Pump Tube Set, PP, 47"
Cole-Parmer Drum Pump Tube Set, SS, 39"
Economical drum pump tube set, 40", PP, Alloy 625 shaft.
More Details
Stainless Steel Fittings
Buy Stainless Steel Fittings Items Including:
Stainless Steel fittings; 0.020" bore, cross, ss (YO-01939-60)
Compression To Thread Adapter, Stainless Steel, 6mm OD Tubing X 1/4" N (YO-31528-24)
Compression To Thread Adapter, Stainless Steel, 8mm OD Tubing X 1/4" N (YO-31528-26).
More Details
Buchner Funnel 110mm Dia
Buy Buchner Funnel 110mm Dia Items Including:
High-density polyethylene Buchner funnel, 42.5 mm dia (YO-06128-02)
High-density polyethylene Buchner funnel, 55 mm dia (YO-06128-03)
Polypropylene Buchner funnel, 110 mm dia (YO-06128-05)
Polypropylene Buchner funnel, 150 mm dia (YO-06128-10).
More Details
Kimax Glass Bottles
Buy Kimax Glass Bottles Items Including:
15 mL Kimax Glass Dropping Bottle
125 mL Kimax Glass Dropping Bottle
KIMAX RESERVOIR with BOTTOM HOSE OUTLET and NARROW MOUTH FOR STOPPER, (YO-34541-01)
KIMAX RESERVOIR with BOTTOM HOSE OUTLET and NARROW MOUTH FOR STOPPER, (YO-34541-03).
More Details
Precision Scientific Water Bath
Buy Precision Scientific Water Bath Items Including:
Precision™ General-Purpose Heated Analog Bath; 1.5 Liters; 115V (YO-12418-00)
Precision™ General-Purpose Heated Analog Bath; 1.5 Liters; 230V (YO-12418-05)
Precision™ General-Purpose Heated Analog Bath; 2.5 Liters; 115V (YO-12418-10).
More Details
Cole-Parmer Glass Stir Rods
Buy Cole-Parmer Glass Stir Rods Items Including:
Cole-Parmer Glass Stir Rods with Rubber Policeman, 10" 12/pk (YO-06369-56)
Cole-Parmer Glass Stir Rods with Rubber Policeman, 12" 12/pk (YO-06369-58)
Stir Rod, Glass, 150mm, 200/pk (40500-150)
Stir Rod, Glass, 250mm, 100/pk (40500-250).
More Details
Luer Lock Stopcock
Buy Luer Lock Stopcock Items Including:
Large-bore 3-way, male-lock, stopcocks, 10/pack (YO-30600-23)
3-way male lock stopcock, 100/pack (YO-30600-25)
adapter; female luer lock to two-way valve; Kynar body and PP insert
Stopcocks with Luer Connections; 1-way; male lock (YO-30600-00).
More Details
Laboratory Mill
Buy Laboratory Mill Items Including:
One-tier jar high-capacity laboratory jar mill; 13" roller, 115 VAC/60 (YO-04149-00)
One-tier jar high-capacity laboratory jar mill; 13" roller, 230 VAC/50 (YO-04149-05).
More Details
Parker Check Valves
Buy Parker Check Valves Items Including:
CHECK VALVE, ACETAL BODY, push-to-connect, 1/4'' tube OD (A4VC4-MG)
CHECK VALVE, ACETAL BODY, push-to connect, 5/16'' tube OD (A5VC5-MG).
More Details
Ptfe Fittings
Buy Ptfe Fittings Items Including:
PTFE Fittings, Cone1/8" OD4/pack (YO-06471-62)
PTFE Fittings, Replacement O-rings, 2 to 4 mm, 10/pack (YO-06473-26)
PTFE Fittings, Grippers, 1/16" size, 10/pack (YO-06473-60)
PTFE Fittings, Grippers, 1/8" size, 10/pack (YO-06473-65).
More Details
Manual Override Solenoid Valve
Buy Manual Override Solenoid Valve Items Including:
Two-Way, Pilot-Operated Valve with Manual Override, 1/2" NPT(F), 120 V (453349M)
Two-Way, Pilot-Operated Valve with Manual Override, 3/4" NPT(F), 120 V (453350J)
Two-Way, Pilot-Operated Valve with Manual Override, 1-1/2" NPT(F), 230.
More Details
Plastic Quick Disconnect Fittings
Buy Plastic Quick Disconnect Fittings Items Including:
Quick-disconnect fittings, Polysulfone fittings, 1/4" ID, body, 5/pack (MPC17004T39)
Quick-disconnect fittings, Polysulfone fittings, 3/8" ID, body, 5/pack (MPC17006T39).
More Details
Tube Furnace
Buy Tube Furnace Items Including:
Horizontal Tube Furnace, 18-7/8" Tube Length, 1500°C, 208 / 240 V (RHTC 80-230/15/P330)
Horizontal Tube Furnace, 24-3/4" Tube Length, 1500°C, 208 / 240 V (RHTC 80-450/15/P330).
More Details
Scalpel Blades and Handles
Buy Scalpel Blades and Handles Items Including:
Siegel scalpel blade handle (YO-10922-20)
Scalpel blade handle, #3, Stainless Steel (YO-10922-22)
Scalpel blade handle, #4, Stainless steel (YO-10922-26)
Scalpel blade handle, #4, Nickel-plated (YO-10922-28)
Scalpel blade handle, #6 (YO-10922-32).
More Details
Ysi Conductivity Solutions
Buy Ysi Conductivity Solutions Items Including:
YSI conductivity calibration standard, 10,000 µS
YSI conductivity calibration standard, 100,000 µS.
More Details
Pressure Gauge for Hydraulic Press
Buy Pressure Gauge for Hydraulic Press Items Including:
Two-Gauge Manifold for Manual Hydraulic Press Test System 59620-00.
More Details
Cimarec Hot Plates
Buy Cimarec Hot Plates Items Including:
Thermo Scientific Cimarec ceramic stirring hot plate, 4x4 120v
Thermo Scientific Cimarec ceramic stirring hot plate, 7 (SP131325Q)
Thermo Scientific Cimarec ceramic stirring hot plate, 10x10 120v
Thermo Scientific Cimarec 4x4 Ceramic Hotplate 120V.
More Details
Cole-Parmer Rotronic Water Activity Meter Systems
Buy Cole-Parmer Rotronic Water Activity Meter Systems Items Including:
Handheld Meter Kit, 14-mm sample cup size, one probe input (one probe (HYGROPALM-AW1 W/14ML CUP).
More Details
Gloss Meters
Buy Gloss Meters Items Including:
High-resolution digital gloss meter
Gloss meter, 20°, 0 to 2000 GU
Gloss meter, 60°, 0 to 2000 GU
Gloss meter, 85°, 0 to 2000 GU
Gloss meter, 20/60/85°, 0 to 2000 GU
More Details
Em Quant Test Strips
Buy Em Quant Test Strips Items Including:
EM Quant Ammonium Test Strips
EM Quant Chloride Test Strips
EM Quant Cyanide Test Strips
EM Quant Lead Test Strips
EM Quant Nickel Test Strips
More Details
Kimble Disposable Culture Tubes
Buy Kimble Disposable Culture Tubes Items Including:
Kimble Disposable Round Bottom Culture Tubes, 13 x 100, no markings, 1
Kimble Disposable Round Bottom Culture Tubes, 16 x 100, no markings, 1
Kimble Disposable Round Bottom Culture Tubes, 16 x 125, no markings, 1.
More Details
100% Nylon Knit Glove Liners
Buy 100% Nylon Knit Glove Liners Items Including:
Glove liners, full-fingered, small, 12 pairs/pk
Glove liners, full-fingered, medium, 12 pairs/pk
Glove liners, full-fingered, large, 12 pairs/pk
Glove liners, full-fingered, x-large, 12 pairs/pk
Glove liners, half-fingered, medium, 12 pairs/pk.
More Details
Nylon Filter Bags
Buy Nylon Filter Bags Items Including:
Nylon monofilament filter bag; 10"L, 100 µm (255009-03)
Nylon monofilament filter bag; 10"L, 200 µm (255011-03)
Nylon monofilament filter bag; 10"L, 250 µm (BN-410-250)
Nylon monofilament filter bag; 18-1/2"L, 50 µm (255255-03).
More Details
A&D Weighing Hc-I Counting Scales
Buy A&D Weighing Hc-I Counting Scales Items Including:
A&D HC-i Counting Scale, 3 kg/6 lb
A&D HC-i Counting Scale, 15 lb x 0.002 lb
A&D HC-i Counting Scale, 30 lb x 0.005 lb
A&D HC-i Counting Scale, 60 x 0.01 lb.
More Details
Hvac/R Accessories for Heat Sealer
Buy Hvac/R Accessories for Heat Sealer Items Including:
Replacement heating elements for heat sealer, 8", 115 VAC (K200H)
Replacement heating elements for heat sealer, 8", 230 VAC (K205H)
Replacement heating elements for heat sealer, 12", 115 VAC (K300H).
More Details
Welding Fume Hoods
Buy Welding Fume Hoods Items Including:
Air Flow Monitor for Fume Hoods, 30 to 400 FPM Monitoring Range
Primary Hydrocarbon Filters for 30" Self-Monitoring Ductless Fume Hood (YO-33713-50)
Primary Hydrocarbon Filters for 54" Self-Monitoring Ductless Fume Hood (YO-33713-52).
More Details
70618
results
<<<Previous
7
8
9
10
11
12
13
14
15
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
25 Ml Volumetric Flask to Welding Fume Hoods
>
31125329-s-toc_2 31125329stoc2