Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 13
Particle Counter Calibration Services
to
Nalgene Fep Wash Bottle
Image
Product Detail
More Detail
Particle Counter Calibration Services
Buy Particle Counter Calibration Services Items Including:
NIST-Traceable Calibration; Counters, High-Accuracy (YO-17101-24).
More Details
Interscience Bagmixer Paddle Lab Blender
Buy Interscience Bagmixer Paddle Lab Blender Items Including:
Paddle Blender; 5 To 100 mL, Variable Speed, Window Door, Removable Pa (YO-36850-09)
Paddle Blender; 50 To 400 mL, Fixed Speed, Stainless Steel Door (YO-36850-10)
Paddle Blender; 50 To 400 mL, Variable Speed, Window Door, Removable P (YO-36850-13).
More Details
Delrin Acetal Manifolds
Buy Delrin Acetal Manifolds Items Including:
Acetal manifold, 2 port 1/4" NPT(F) inlet, 1/8" NPT(F) outlet (YO-31524-00)
Acetal manifold, 2 port 3/8" NPT(F) inlet, 1/4" NPT(F) outlet (YO-31524-02).
More Details
Sanitary Fittings
Buy Sanitary Fittings Items Including:
Nylon Clamp for Sanitary Fittings, 1" to 1-1/2" (YO-30508-23)
Nylon Clamp for Sanitary Fittings, 2" (YO-30508-25)
Nylon Clamp for Sanitary Fittings, 3" (YO-30508-27)
Nylon Clamp for Sanitary Fittings, 4" (YO-30508-29).
More Details
Gilmont Micrometer Burettes
Buy Gilmont Micrometer Burettes Items Including:
Gilmont micrometer burette, 2 mL
Gilmont micrometer burette tip, 2 mL, for use with 07846-00
Gilmont Micrometer Syringe, 0.2 mL
Gilmont micrometer syringe tip, for use with 07840-00
Gilmont micrometer syringe, 2 mL.
More Details
1 Male Pvc Threaded Plug
Buy 1 Male Pvc Threaded Plug Items Including:
Threaded plug, polyethylene, 1/2" male NPT, 10/pack (YO-30401-39)
Threaded plug, polyethylene, 1/2" male NPT, 100/pack (YO-30401-41)
Threaded plug, polyethylene, 1/4" male NPT, 100/pack (YO-30401-45).
More Details
1 Brass Barb Fitting to Npt
Buy 1 Brass Barb Fitting to Npt Items Including:
Barbed fittings, NPT male tees, Natural PP, 1" NPT x 1" ID, 25/32", (YO-30610-89)
Barbed fittings, NPT male tees, Natural PP, 1/2" NPT x 1/4" ID, 1/8", (YO-30640-80).
More Details
Cole-Parmer Natural Polypropylene Straight Barbed Connectors
Buy Cole-Parmer Natural Polypropylene Straight Barbed Connectors Items Including:
Straight barbed union, polypropylene, 3/16" ID, 10/pack (YO-30622-57)
Barbed fittings, Straight Connector, Natural PP, 1" ID, 25/32", 4", 1- (YO-30610-06)
Straight Connector, Polypropylene, 3/32", 100 PACK (YO-30626-46).
More Details
Reducing Bushings Threaded Fittings
Buy Reducing Bushings Threaded Fittings Items Including:
Reducing bushings, nylon, 1/4 NPT(M) x 1/8 NPT(F), pack of 6 (YO-06349-30)
Reducing bushings, nylon, 3/8 NPT(M) x 1/8 NPT(F), pack of 6 (YO-06349-31)
Reducing bushing (YO-06349-32)
Reducing bushing 1 1/2" x 2" (YO-06445-71).
More Details
Cleanroom Chairs
Buy Cleanroom Chairs Items Including:
Cleanroom Chair, ESD, 19.5" to 27" height, aluminum base
Cleanroom Chair, ESD Polyurethane, 16" to 21" height, aluminum base
Clean Room Vinyl Chair, 19" - 24" Height
Clean Room Vinyl Chair, 24" - 29" Height.
More Details
Temperature Indicating Labels
Buy Temperature Indicating Labels Items Including:
Five-Point temperature indicating labels; temperature points 105-130d (YO-90303-20)
Five-Point temperature indicating labels; temperature points 140-180d (YO-90303-22)
Descending Irreversible Low-Temperature Indicators, 32°F (0°C) (YO-90309-60).
More Details
Kimax Glassware 2l Graduated Cylinder
Buy Kimax Glassware 2l Graduated Cylinder Items Including:
100 mL Kimax class B graduated cylinders
250 mL Kimax class B graduated cylinders
500 mL Kimax class B graduated cylinders
1000 mL Kimax class B graduated cylinders
2000 mL Kimax class B graduated cylinders
More Details
Thermo Scientific Nalgene Polypropylene Bottles
Buy Thermo Scientific Nalgene Polypropylene Bottles Items Including:
Thermo Scientific Nalgene polypropylene jar, 2 1/4 quart
Thermo Scientific Nalgene polypropylene jar, 4 7/8 quart
Thermo Scientific Nalgene polypropylene narrow-mouth bottle, 1/8 oz
Thermo Scientific Nalgene polypropylene narrow-mouth bottle, 1 oz.
More Details
Hach Conductivity/Resistivity/Tds Handheld Meters
Buy Hach Conductivity/Resistivity/Tds Handheld Meters Items Including:
Hach® sensION™ 5 Waterproof Conductivity Meter
Hach SensION™+ Rugged Conductivity Meter Kit
Hach SensION™+ Conductivity Meter Kit
Cond
Cell, General.
More Details
Eppendorf Series 2000 Reference Pipettors
Buy Eppendorf Series 2000 Reference Pipettors Items Including:
Fixed-volume pipettors, 2000 µL (22 47 175-9)
Ultramicro Fixed-Volume Pipettors, 1 µL (22 47 040-0)
Ultramicro Fixed-Volume Pipettors, 10 µL (22 47 055-8)
Fixed-volume pipettors, 10 µL (22 47 060-4)
Fixed-volume pipettors, 100 µL (22 47 115-5).
More Details
Gas Cylinder Stands
Buy Gas Cylinder Stands Items Including:
Stand (970040)
Portable Stand
PVC Coated Stand.
More Details
Buchner Funnels
Buy Buchner Funnels Items Including:
High-density polyethylene Buchner funnel, 42.5 mm dia (YO-06128-02)
High-density polyethylene Buchner funnel, 55 mm dia (YO-06128-03)
Polypropylene Buchner funnel, 90 mm dia (YO-06128-04)
COORSTEK Buchner Funnels, Ceramic, 2 mL, 1/pk (YO-17815-02).
More Details
316 Stainless Steel Barbed Cross Connectors
Buy 316 Stainless Steel Barbed Cross Connectors Items Including:
Barbed Fittings; Cross Connector, 316 SS, 1/16" Tubing ID (YO-31208-51)
Barbed Fittings; Cross Connector, 316 SS, 3/32" Tubing ID (YO-31208-52)
Barbed Fittings; Cross Connector, 316 SS, 3/16" Tubing ID (YO-31208-54).
More Details
Accumet Microprobe Combination Ph Electrodes
Buy Accumet Microprobe Combination Ph Electrodes Items Including:
Accumet MicroProbe™ combination electrode, rigid stem 1-1/2" L
Accumet pH Electrode, Combination, Microprobe, Refillable, Glass Body,
Accumet pH Electrode, Combination, Microprobe, Long-neck, Refillable,.
More Details
Laboratory Wash Bottles
Buy Laboratory Wash Bottles Items Including:
Thermo Scientific Nalgene "Right-to-Know" safety wash bottle for ethan (2425-0502)
Thermo Scientific Nalgene "Right-to-Know" safety wash bottle for ethan (2425-1002)
Thermo Scientific Nalgene fluorinated wash bottle, 500 mL.
More Details
Corning Pyrex Vista Glass Beakers
Buy Corning Pyrex Vista Glass Beakers Items Including:
Pyrex Vista Griffin Style Beakers, 100mL 12/pk (70000-100)
Pyrex Vista Griffin Style Beakers, 150mL 12/pk (70000-150)
Pyrex Vista Griffin Style Beakers, 250mL 12/pk (70000-250)
Pyrex Vista Griffin Style Beakers, 400mL 12/pk (70000-400).
More Details
Diamond Grip Latex Gloves
Buy Diamond Grip Latex Gloves Items Including:
Microflex Diamond Grip™ Latex Gloves; size, small
Microflex Diamond Grip™ Latex Gloves; size, medium
Microflex Diamond Grip™ Latex Gloves; size, extra-large
Cole-Parmer Raised Diamond Grip Nitrile Gloves, single-layer, light we (YO-40216-02).
More Details
Stainless Steel Beakers with Handles
Buy Stainless Steel Beakers with Handles Items Including:
Stainless-steel Griffin-style beaker with easy-pour rim, 125 mL (YO-07205-30)
Stainless-steel Griffin-style beaker with easy-pour rim, 250 mL (YO-07205-40)
Stainless steel Griffin-style beaker with easy-pour rim, 600 mL (YO-07205-50).
More Details
Lamotte Turbidity in Water Test Kit
Buy Lamotte Turbidity in Water Test Kit Items Including:
Watersafe Well Water Test Kit
Soil texture test kit (1067)
Garden test kit (5679)
Soil Nutrient Test Kit; Macronutrient and pH (5029)
LaMotte Dissolved Oxygen Test Kit, 50 tests/kit
More Details
Thermo Scientific Nalgene Separatory Funnels
Buy Thermo Scientific Nalgene Separatory Funnels Items Including:
Thermo Scientific Nalgene separatory funnel with polypropylene body an (YO-06126-10)
Thermo Scientific Nalgene separatory funnel with polypropylene body an (YO-06126-20)
Thermo Scientific Nalgene Funnel, separatory, FEP, 1000mL, 1/pk.
More Details
Cole-Parmer One-Way Luer Check Valves
Buy Cole-Parmer One-Way Luer Check Valves Items Including:
One-way luer check valve, SAN/LDPE with silicone diaphragm, female-mal (SCV08053).
More Details
Grinding Media
Buy Grinding Media Items Including:
Zirconia grinding media, 3/8" x 3/8" (YO-04158-10)
Zirconia grinding media, 1/2" x 1/2" (YO-04158-20)
Burundum grinding media, 1/2" x 1/2" (YO-04150-10)
Burundum grinding media, 13/16" x 13/16" (YO-04150-20).
More Details
Flammable Storage Cabinets
Buy Flammable Storage Cabinets Items Including:
SciMatCo Flammable Storage Cabinet, 4 gallon capacity, self-closing do
SciMatCo Flammable Storage Cabinet, 22 gallon capacity, self-closing d
Flammable Storage Cabinet, Manual-Latching Doors, 90 Gallon (YO-09418-29).
More Details
Advantec Quartz Fiber Filter Sheets
Buy Advantec Quartz Fiber Filter Sheets Items Including:
Advantec Grade QR100 Quartz Fiber Filters, 25-mm dia; box of 100.
Advantec Grade QR200 Quartz Fiber Filters, 25-mm dia; box of 100.
Advantec Grade QR200 Quartz Fiber Filters, 47-mm dia; box of 50.
Quartz fiber filter, QR100, 55-mm dia; box of 100..
More Details
Nalgene Fep Wash Bottle
Buy Nalgene Fep Wash Bottle Items Including:
Thermo Scientific Nalgene Wash bottle, FEP, 125mL, 1/pk
Thermo Scientific Nalgene Wash bottle, FEP, 250mL, 1/pk
Thermo Scientific Nalgene Wash bottle, FEP, 500mL, 1/pk.
More Details
70618
results
<<<Previous
9
10
11
12
13
14
15
16
17
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Particle Counter Calibration Services to Nalgene Fep Wash Bottle
>
31125329-s-toc_2 31125329stoc2