Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 14
Nanopure Infinity Water Purification Systems Cartridges
to
Electrode Storage Solution
Image
Product Detail
More Detail
Nanopure Infinity Water Purification Systems Cartridges
Buy Nanopure Infinity Water Purification Systems Cartridges Items Including:
Thermo Scientific Barnstead Pretreatment Cartridge for reverse osmosis (D50250)
Thermo Scientific Barnstead Pretreatment Cartridge for tap and deioniz (D50251)
Thermo Scientific Barnstead Ultrapure deionization cartridge, 680 grai (D50253).
More Details
Metal Quick Disconnect Fittings
Buy Metal Quick Disconnect Fittings Items Including:
Barbed fittings, Metal quick-disconnect fittings, coupling bodies,valv (LCD13006)
Barbed fittings, Metal quick-disconnect fittings, coupling bodies,valv (LCD17005)
Barbed fittings, Metal quick-disconnect fittings, coupling bodies,valv (LCD17006).
More Details
Guaranteed I Water Quality Test Strips
Buy Guaranteed I Water Quality Test Strips Items Including:
Water Quality Test Strips, Multi-test strip
(480115)
Iron test strips (480025)
Total Hardness Test Strips (480008)
Free chlorine test strips (481026)
More Details
Spirax Sarco Steam Traps
Buy Spirax Sarco Steam Traps Items Including:
Thermo-Dynamic steam trap, 3/8"NPT(F) (TD52 3/8" 54529C)
Cast iron inverted bucket steam trap; 3/4" NPT(F), 75 psig (B1H-75.75 in 64161 )
Cast iron inverted bucket steam trap; 3/4" NPT(F), 125 psig (B1H-125.75 in 64162 ).
More Details
Ammonium Hydroxide Chemical Formula
Buy Ammonium Hydroxide Chemical Formula Items Including:
AMMONIUM HYDROXIDE ACS 2.5L (YO-88053-91)
AMMONIUM HYDROXIDE 25% 2.5LT (YO-88252-97)
AMMONIUM HYDROXIDE REAG 2.5L (YO-88360-61).
More Details
Rotronic Water Activity Meter Systems
Buy Rotronic Water Activity Meter Systems Items Including:
Handheld Meter Kit, 14-mm sample cup size, one probe input (one probe (HYGROPALM-AW1 W/14ML CUP).
More Details
Lmi Manually Adjustable Control Metering Pumps
Buy Lmi Manually Adjustable Control Metering Pumps Items Including:
Manual control solenoid diaphragm metering pump, 2.5 GPH, 115 VAC
Manual control solenoid diaphragm metering pump, 0.42 GPH, 115 VAC
Manual control solenoid diaphragm metering pump, 0.42 GPH, 230 VAC.
More Details
Safety First Aid Station (Symbol) Signs
Buy Safety First Aid Station (Symbol) Signs Items Including:
Safety Sign, First Aid Station (symbol), 7" X 10", Adhesive Vinyl
Safety Sign, First Aid Station (symbol), 7" X 10", Aluminum
Safety Sign, First Aid Station (symbol), 10" X 14", Adhesive Vinyl.
More Details
Myron L Conductivity/Resistivity/Tds Handheld Meters
Buy Myron L Conductivity/Resistivity/Tds Handheld Meters Items Including:
MYRON L 512T5 analog TDS meter
MYRON L 532T1 analog TDS meter
MYRON L 512M5 analog conductivity meter
MYRON L 532M1 analog conductivity meter
MYRON L EP-10 analog conductivity meter
More Details
Diaphragm Metering Pumps
Buy Diaphragm Metering Pumps Items Including:
Programmable Remote Control Solenoid-driven Diaphragm Metering Pump wi (EHE30E1-PC)
Programmable Remote Control Solenoid-driven Diaphragm Metering Pump wi (EHE30E2-PC)
Manual control solenoid diaphragm metering pump, 2.5 GPH, 115 VAC.
More Details
Glass Funnel
Buy Glass Funnel Items Including:
ACE GLASS Separatory Funnels, 125 mL (YO-34731-02)
ACE GLASS Separatory Funnels, 1000 mL (YO-34731-08)
ACE GLASS Separatory Funnels, 2000 mL (YO-34731-10)
Funnel, Ace, Powder Offset, 75MM (YO-34732-00)
Weighing Funnels, Glass, Micro (YO-34700-00).
More Details
Calibrated Graduated Cylinders
Buy Calibrated Graduated Cylinders Items Including:
Thermo Scientific Nalgene calibrated graduated polymethylpentene cylin (3663-0010)
Thermo Scientific Nalgene calibrated graduated polymethylpentene cylin (3663-0025)
Corning Graduated Cylinder 10ml PP, 2/pk
Corning Graduated Cylinder 50ml PP, 2/pk.
More Details
Sartorius Balances
Buy Sartorius Balances Items Including:
RS-232 Adapter for Sartorius Element balances
USB Adapter for Sartorius Element balances
Sartorius Talent Portable Balances, 400g 115VAC (TE412)
Sartorius Talent Portable Balances, 500g 115VAC (TE612)
Sartorius Element™ 100 g x 0.001 g (ELT103.1).
More Details
Luer Lock Fittings Hypo Tube Clamping
Buy Luer Lock Fittings Hypo Tube Clamping Items Including:
Rotating female luer to male luer lock adapter, Nickel-plated brass, e (YO-31507-80)
Luer fittings, 316 SS fittings, female luer lock x 1/16" hose barb (YO-31507-29)
Luer fittings, Chrome-plated Luer adapters,male luer lock plug (YO-31507-52).
More Details
Genova Nano Micro-Volume Uv/Visible Spectrophotometer
Buy Genova Nano Micro-Volume Uv/Visible Spectrophotometer Items Including:
GENOVA NANO Life Science Analyzer for DNA, RNA, Oligo, Protein analysi
Micro-Cuvette Holder with reduced aperature for use with non-masked mi
8 Cell Automatic Turet
Sipper Pump
Sipper -Peltier
More Details
High-Temperature Industrial Mechanical Convection Ovens
Buy High-Temperature Industrial Mechanical Convection Ovens Items Including:
High-temperature industrial mechanical convection oven, 4.3 cu ft, 115
Industrial Mechanical Convection Ovens, Drawer oven, 4.8 cu ft, 115 VA
Industrial Mechanical Convection Ovens, Drawer oven, 4.8 cu ft 230 VAC.
More Details
Rotary Evaporators
Buy Rotary Evaporators Items Including:
IKA Basic Rotary Evaporator With Vertical Uncoated Glassware, 115 VAC
IKA Basic Rotary Evaporator With Vertical Safety-coated Glassware 115V (YO-28710-05)
IKA Basic Rotary Evaporator With Vertical Safety Coated Glassware 230V (YO-28710-07).
More Details
Dickson Chart Recorders
Buy Dickson Chart Recorders Items Including:
Dickson PW470 Pressure Chart Recorder, 0 to 100 PSI
Dickson PW474 Pressure Chart Recorder, 0 to 200 PSI
Chart Paper for 8" Circular Recorders; 24 hour, 20 to 120°, 60 she
Chart Paper for 8" Circular Recorders; 24 hour, 20 to 50°, 60 shee.
More Details
Lamotte 5860 Dissolved Oxygen Test Kit
Buy Lamotte 5860 Dissolved Oxygen Test Kit Items Including:
LaMotte Dissolved Oxygen Test Kit Refill; Winkler Titration Method, 50.
More Details
Luer Lock Male to Male Stopcock
Buy Luer Lock Male to Male Stopcock Items Including:
Stopcocks with Luer Connections; 3-way; male lock; 5 flow pattern; 100 (YO-30600-07)
Stopcocks with Luer Connections; 4-way; male lock; 100/pack (YO-30600-09)
Luer stopcock, 1-way, male luer lock x male luer lock, black PVDF, pac (YO-31200-90).
More Details
Sanitary Pressure Gauge
Buy Sanitary Pressure Gauge Items Including:
Sanitary Pressure Gauge, 0-30 PSI, 3/4
Sanitary Pressure Gauge, 0-60 PSI, 3/4
Sanitary Pressure Gauge, 0-100 PSI, 3/4 Tri-clamp
Sanitary Pressure Gauge, 0-160 PSI, 3/4
Digital Sanitary Gauge, 1.5" Tri-Clamp, 30 to 0" Hg, battery powered.
More Details
Test Tube Racks in Atlanta
Buy Test Tube Racks in Atlanta Items Including:
Scienceware test tube rack for 10- to 13-mm tubes
Scienceware polypropylene test tube rack for 26-30 mm tubes
Thermo Scientific Nalgene Unwire™ test tube rack for 13-mm tubes, w
Thermo Scientific Nalgene Unwire™ test tube rack for 13-mm tubes, o.
More Details
Bronze Globe Valves
Buy Bronze Globe Valves Items Including:
Bronze globe valve; 1" NPT(F) (GLV0700750)
Bronze globe valve; 1-1/4" NPT(F) (GLV0700760)
Bronze globe valve; 1-1/2" NPT(F) (GLV0700770).
More Details
Handheld Ph Meters
Buy Handheld Ph Meters Items Including:
Thermo Scientific Orion 3-Star handheld pH meter kit
Thermo Scientific Orion 3-Star handheld pH meter
Oakton pH meter with probe
Horiba D-51 Waterproof pH Meter
Hach waterproof ISFET pH meter kit
More Details
Stablcal Stabilized Formazin Turbidity Standards
Buy Stablcal Stabilized Formazin Turbidity Standards Items Including:
Hach StablCal 0.1 NTU Turbidity Standard, 500-mL
Hach StablCal 20 NTU Turbidity Standard, 500-mL
Hach StablCal 200 NTU Turbidity Standard, 500-mL
Hach StablCal 800 NTU Turbidity Standard, 500-mL.
More Details
Porcelain Ball Mill Jars
Buy Porcelain Ball Mill Jars Items Including:
Alumina-fortified porcelain milling jar, 0.1 gal
(YO-04160-00)
Alumina-fortified porcelain milling jar, 0.5 gal
(YO-04162-00)
Alumina-fortified porcelain milling jar, 1.5 gal
More Details
Disposable Pipette Tips
Buy Disposable Pipette Tips Items Including:
Cole-Parmer Disposable Pipette Tips, sterile, 2" L, 0-200 µL, 960/c (A23091PRX)
Cole-Parmer Disposable Pipette Tips, sterile, 2" L, 0-200 µL, 960/c (A23071PRX)
Cole-Parmer Disposable Pipette Tips, non-sterile, 3-1/8" L, 200-1300.
More Details
Nylon Mesh Screen Sieve
Buy Nylon Mesh Screen Sieve Items Including:
Mesh Screen, Nylon, 1.70 mm sieve opening, 10 mesh
Mesh Screen, Nylon, 1.40 mm sieve opening, 12 mesh
Mesh Screen, Nylon, 1.18 mm sieve opening, 14 mesh
Mesh Screen, Nylon, 0.850 mm sieve opening, 20 mesh.
More Details
Sanitary Eccentric Reducer
Buy Sanitary Eccentric Reducer Items Including:
Polypropylene Sanitary Eccentric Reducer, 1 1/2" x 3/4" Mini Clamp Si (YO-31805-01)
Cole-Parmer, Eccentric Sanitary Clamp Reducer, 304L SS, 1.5" X 1"
ECCENTRIC SANITARY CLAMP REDUCER, 316L SS, 1-1/2" X 1" (YO-30560-05).
More Details
Electrode Storage Solution
Buy Electrode Storage Solution Items Including:
Thermo Scientific Orion pH Electrode Storage Solution, 1 pint (475-mL)
Thermo Scientific Orion pH Electrode Storage Solution, 5 x 60-mL bottl
Oakton electrode cleaning solution, case of 24
Thermo Scientific Orion pH Electrode Cleaning Solution Kit.
More Details
70618
results
<<<Previous
10
11
12
13
14
15
16
17
18
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Nanopure Infinity Water Purification Systems Cartridges to Electrode Storage Solution
>
31125329-s-toc_2 31125329stoc2