Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 15

Extech Light Meters   to   Luer Lock Fittings Iv Tubing

Image
  Product Detail
More Detail

Extech Light Meters

Buy Extech Light Meters Items Including:
  • Extech Heavy-Duty Light Meter, Model HD400
  • Extech Heavy-Duty Datalogging Light Meter, Model HD450
  • Extech EasyView ™ Light Meter with Memory
  • Extech SDL400 Light Meter with datalogging and SD card
  • Extech EasyView ™ Light Meter with NIST Certificate.

Nalgene Autoclavable Wash Bottles

Buy Nalgene Autoclavable Wash Bottles Items Including:
  • Thermo Scientific Nalgene PPCO wash bottle, 500 mL
  • Thermo Scientific Nalgene PPCO wash bottle, 1 L
  • Thermo Scientific Nalgene Wash bottle, FEP, 125mL, 1/pk.

Nalgene Ldpe Wash Bottle

Buy Nalgene Ldpe Wash Bottle Items Including:
  • Thermo Scientific Nalgene economy low-density polyethylene wash bottle (2401-0125)
  • Thermo Scientific Nalgene economy low-density polyethylene wash bottle (2401-0250)
  • Thermo Scientific Nalgene economy low-density polyethylene wash bottle (2401-0500).

Thermo Scientific Heating Immersion Circulators

Buy Thermo Scientific Heating Immersion Circulators Items Including:
  • Thermo Scientific Immersion Circulator w/bridge, AC 150 thermostat (23
  • Thermo Scientific Immersion Circulator, AC 200 thermostat with bridge (AC 200) (YO-12135-40)
  • Thermo Scientific Immersion Circulator, PC 300 thermostat with bridge.

L/S Multichannel Pump Heads

Buy L/S Multichannel Pump Heads Items Including:
  • L/S 4-channel multichannel pump head for microbore tubing (YO-07534-04)
  • L/S 8-channel multichannel pump head for microbore tubing (YO-07534-08)
  • L/S 4-channel multichannel pump head for L/S tubing
  • L/S 8-channel multichannel pump head for L/S tubing.

Centrifuge Tube Filters

Buy Centrifuge Tube Filters Items Including:
  • Costar Spin-X Centrifuge Tube Filters, Spinfilter Ca 0.22um 100/cs
  • Costar Spin-X Centrifuge Tube Filters, Spinfilter Ca 0.45µm 100/cs
  • Costar Spin-X Centrifuge Tube Filters, Spinfilter Ny 0.22um 200/cs.

Small Batch Mixer

Buy Small Batch Mixer Items Including:
  • Lightnin small batch mixer with two-speed motor, 115 VAC
  • Lightnin small batch, fixed-speed mixer with 1/20-hp explosion-proof m.

Stirring Hot Plates

Buy Stirring Hot Plates Items Including:
  • Cole-Parmer Five-Position Stirring Hot Plate, 120 VAC
  • Cole-Parmer Nine-Position Stirring Hot Plate, 120 VAC
  • Cole-Parmer Five-Position Stirring Hot Plate, 230 VAC
  • EchoTherm™ Digital Stirring Hot Plate , 115 VAC
  • Basic Mini Stirring Hot Plate, 120V.

Male Luer Tube Fittings

Buy Male Luer Tube Fittings Items Including:
  • 316 SS fittings; female luer x male luer lock (YO-31507-21)
  • 316 SS fittings; male luer lock plug (YO-31507-23)
  • 316 SS fittings; male luer lock x 10-32 UNF thread (YO-31507-31)
  • 316 SS fittings; male luer lock x 1/4-28 UNF thread (YO-31507-32).

Stainless Steel Bulkhead Fitting

Buy Stainless Steel Bulkhead Fitting Items Including:
  • Compression To Thread Adapter, Stainless Steel, 6mm OD Tubing X 1/4" N (YO-31528-24)
  • Compression Straight Union, Stainless Steel, 10mm OD Tubing (YO-31528-35)
  • Compression Reducing Union, Stainless Steel, 12mm X 6mm OD Tubing (YO-31528-41).

Em Quant Water Testing

Buy Em Quant Water Testing Items Including:
  • EM Quant Ammonium Test Strips
  • EM Quant Chloride Test Strips
  • EM Quant Cyanide Test Strips
  • EM Quant Lead Test Strips
  • EM Quant Nickel Test Strips

Cole-Parmer 3-Way Solenoid Pinch Valves

Buy Cole-Parmer 3-Way Solenoid Pinch Valves Items Including:
  • Solenoid-Operated Two-Way Pinch Valve; Normally-Closed, 10 mm Tube OD, (PK-0802-NC-3 12V N CLOSED)
  • Solenoid-Operated Three-Way Pinch Valve, 12 VDC (PM-0815W 12VDC N OPEN)
  • Solenoid-Operated Three-Way Pinch Valve, 24 VDC (PM 0815W, 24VDC N OPEN).

Revco Refrigerator

Buy Revco Refrigerator Items Including:
  • Laboratory undercounter refrigerator (single door); 5.4 CU
  • FT; 115V (REL-404A)
  • Laboratory space-saver refrigerator (single door); 11.5 CU FT; 115V (REL-1204A-20)
  • Laboratory space-saver refrigerator (single door); 11.5 CU FT; 200/230 (REL-1204D).

Check Valve 3 4 Inch

Buy Check Valve 3 4 Inch Items Including:
  • Bel-Art PP Check Valve, 1/4" to 3/8" hose barb, 6/bag
  • PVC Ball Check Valve, 3/8" NPT(F) (161.360.581)
  • PVDF Miniature Check Valve, 3/16" hose barb, Viton diaphragm
  • PVDF Miniature Check Valve, 3/8" hose barb, Viton diaphragm.

Ansell Edmont Gloves

Buy Ansell Edmont Gloves Items Including:
  • Ansell Edmont Sterile Latex Surgical Gloves; size 8 (870 398706)
  • Ansell Edmont Sol-Vex Nitrile Gloves; Unlined; size 10; 11-mil; 13"L; (37-185 117301)
  • Ansell Edmont Sol-Vex Nitrile Gloves; Unlined; size 10; 11-mil; 13"L; (37-145 117076).

Male Luer Fitting

Buy Male Luer Fitting Items Including:
  • Adapters with 1/4-28 UNF thread; male luer fitting; ETFE (YO-06471-90)
  • Adapters with caps; male luer fitting; Tefzel
  • Male luer x male luer adapter, PC, 25/pk (YO-45504-72)
  • Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55).

Honeywell Chart Recorders

Buy Honeywell Chart Recorders Items Including:
  • Replacement Chart Paper for Honeywell DR4300 10" Process Recorders, 10
  • Thermal Paper for Honeywell DR4500A 12" Digital Recorders
  • Honeywell DR4300 10" Process Recorder, 1 channel
  • Honeywell DR4300 10" Process Recorder, 2 channels.

Sanitary Stainless Steel Barbed Fittings

Buy Sanitary Stainless Steel Barbed Fittings Items Including:
  • Cole-Parmer, Barbed sanitary clamp adapter, 316L SS, 3/4-inch clamp x (YO-30700-58)
  • Cole-Parmer, Barbed sanitary clamp adapter, 316L SS, 3/4-inch clamp x (YO-30700-59)
  • Cole-Parmer, Barbed sanitary clamp adapter, 316L SS, 1.5" clamp x 1.5".

Ceramic Buchner Funnel Table Top

Buy Ceramic Buchner Funnel Table Top Items Including:
  • COORSTEK Buchner Funnels, Ceramic, 550 mL, 1/pk (YO-17815-12)
  • COORSTEK Buchner Funnels, Ceramic, 700 mL, 1/pk (YO-17815-14)
  • COORSTEK Buchner Funnels, Ceramic, 30 mL, 12/cs (YO-17815-24)
  • COORSTEK Buchner Funnels, Ceramic, 320 mL, 4/cs (YO-17815-30).

Npt to 1 4 Hose Barb

Buy Npt to 1 4 Hose Barb Items Including:
  • Barbed fittings, NPT male pipe adapter, Natural PP, 1/4" NPT x 1/4" I (YO-30610-38)
  • Barbed fittings, NPT male branch tees, Natural PP, 1/8" NPT x 1/4" ID, (YO-30610-72).

Thermo Scientific Fume Hoods

Buy Thermo Scientific Fume Hoods Items Including:
  • Standing Height Wall Assembly with Fume Hood - 13ft (YO-41524-50)
  • Complete 4foot Fume Hood System With Acid Storage Cabinet Epoxy Worksu (YO-41525-00)
  • Complete 4foot Fume Hood System With General Base Storage Cabinet Epox (YO-41525-05).

Female Compression to Tube Adapter

Buy Female Compression to Tube Adapter Items Including:
  • Compression threaded adapter, PFA, 1/8" tubing OD x 1/8" female NPT (YO-31320-50)
  • Compression threaded adapter, PFA, 1/8" tubing OD x 1/4" female NPT (YO-31320-51).

To 1 16 Id Barb Fitting

Buy To 1 16 Id Barb Fitting Items Including:
  • Barbed fittings, T connector, Kynar, 1/16" ID, 1/32", 1/2", 5/16"; 10/ (YO-30703-70)
  • Barbed fittings, T connector, Nylon, 1/16" ID, 1/32", 25/32", 25/32"; (YO-31500-30).

100 Ml Graduated Cylinder

Buy 100 Ml Graduated Cylinder Items Including:
  • Cylinder, Pyrex Brand 3025, graduated, 100 mL
  • Cylinder, Pyrex Brand 3025, graduated, 100 mL, case of 12
  • Pyrex Brand 3024 cylinder; 100 mL, case of 12
  • Pyrex Brand 3042 cylinder; 100 mL, case of 24
  • Pyrexplus Brand 63024 cylinder; 100 mL, pack of 1.

Ysi Model 550a Dissolved Oxygen Meter

Buy Ysi Model 550a Dissolved Oxygen Meter Items Including:
  • YSI Model 550A Dissolved Oxygen Meter, 25-ft (7.6-m) probe cables.

600 Ml Low Form Griffin Beaker

Buy 600 Ml Low Form Griffin Beaker Items Including:
  • Pyrex Brand 1000 Griffin Low-Form Beaker, 600 mL, 6/pk
  • Pyrex Brand 1000 Griffin Low-Form Beaker, 600 mL, 36/cs.

200 Ml Graduated Glass Centrifuge Tubes

Buy 200 Ml Graduated Glass Centrifuge Tubes Items Including:
  • Conical-Bottom Glass Centrifuge Tubes, 12 mL, with Graduations; 12/cas (8140-12)
  • Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 10 ml, 12 (99502-10)
  • Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 15 ml, 12 (99502-15).

50 Ml Sterile Glass Centrifuge Tube

Buy 50 Ml Sterile Glass Centrifuge Tube Items Including:
  • Conical-Bottom Glass Centrifuge Tubes, 50 mL, Penny-Head Stopper, 12/c (8084-50)
  • Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 10 ml, 12 (99502-10)
  • Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 15 ml, 12 (99502-15).

British Pipe Thread Fittings

Buy British Pipe Thread Fittings Items Including:
  • Threaded adapter, stainless steel, 1/8" female NPT to 1/8" male BSPP (YO-31830-50)
  • Threaded adapter, stainless steel, 1/4" female NPT to 1/4" male BSPP (YO-31830-51).

Luer Lock Fittings Iv Tubing

Buy Luer Lock Fittings Iv Tubing Items Including:
  • Snap luer lock ring, PP, 25/pk (YO-45503-74)
  • Snap luer lock ring, Nylon, 25/pk (YO-45505-74)
  • Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
  • 316 Stainless Steel fittings, male luer lock x male luer lock (YO-31507-90).



Extech Light Meters to Luer Lock Fittings Iv Tubing

>