Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 16
Whatman Filter Paper Sizes
to
Tirrill Burner
Image
Product Detail
More Detail
Whatman Filter Paper Sizes
Buy Whatman Filter Paper Sizes Items Including:
Whatman Qualitative Filter Papers; 5.5 cm dia; pore size, 20-25µ; 1
Whatman Qualitative Filter Papers; 11.0 cm dia; pore size, 20-25 µ;
Whatman Qualitative Filter Papers; 12.5 cm dia; pore size, 20-25 µ;.
More Details
Cryogenic Gloves
Buy Cryogenic Gloves Items Including:
Water-resistant Cryo-Glove gloves, 12"L, medium, 1 pair
Water-resistant Cryo-Glove gloves, 12"L, large, 1 pair
Cryo-Industrial gloves, mid-arm style, medium, 15" length
Cryo-Industrial gloves, mid-arm style, large, 15" length.
More Details
50 Ml Plastic Sample Vial
Buy 50 Ml Plastic Sample Vial Items Including:
Thermo Scientific Nalgene Sample vials with snap caps, 50 mL.
More Details
Millipore Amicon Ultra Centrifugal Filters
Buy Millipore Amicon Ultra Centrifugal Filters Items Including:
Millipore Amicon Ultra Centrifugal Filters, 4ML 100K 24PK
Millipore Amicon Ultra Centrifugal Filters, 15ML 10K 24PK
Millipore Amicon Ultra Centrifugal Filters, 15ML 100K 24PK
Millipore Amicon Ultra Centrifugal Filters, 0.5 mL, 3K NMWL; pack of 2.
More Details
Microcentrifuge Tubes
Buy Microcentrifuge Tubes Items Including:
Cole-Parmer Microcentrifuge Tube, 0.6mL, natural, non sterile
Cole-Parmer Microcentrifuge Tube, 0.6mL, Amber, non sterile
Cole-Parmer Microcentrifuge Tube, 1.5mL, Natural, non sterile
Cole-Parmer Microcentrifuge Tube, 1.5mL, assorted, non sterile.
More Details
Orion Electrode Storage Solution 810007
Buy Orion Electrode Storage Solution 810007 Items Including:
Thermo Scientific Orion pH Electrode Storage Solution, 1 pint (475-mL)
Thermo Scientific Orion pH Electrode Storage Solution, 5 x 60-mL bottl
Thermo Scientific Orion electrode fill solution, 3 M KCl, 5 x 60mL bot
Oakton electrode storage solution.
More Details
Luer Fitting Syringe Adapter
Buy Luer Fitting Syringe Adapter Items Including:
Luer fittings, Polycarbonate fittings, male lock x male lock, rotating (YO-06464-90)
Luer fittings, Chrome-plated Luer adapters,female luer plug (YO-31507-53)
Luer fittings, Chrome-plated Luer adapters,male luer lock to 1/8" to 3 (YO-31507-61).
More Details
Nalgene Solvent Wash Bottles
Buy Nalgene Solvent Wash Bottles Items Including:
Thermo Scientific Nalgene wide-mouth Unitary wash bottle, 1 L
Thermo Scientific Nalgene "Right-to-Know" safety wash bottle for ethan (2425-0502)
Thermo Scientific Nalgene "Right-to-Know" safety wash bottle for ethan (2425-1002).
More Details
304 Stainless Steel Tubing
Buy 304 Stainless Steel Tubing Items Including:
304 Stainless Steel Tubing, 1/8" OD, 0.085" ID, 50ft roll (YO-34680-50)
BARBED SANITARY CLAMP ADAPTER, 304L SS, 1" CLAMP X 1" ID TUBING (YO-30561-13)
ECCENTRIC SANITARY CLAMP REDUCER, 304L SS, 1-1/2" X 1" (YO-30561-05).
More Details
Ac Power Supplies
Buy Ac Power Supplies Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
Bain Marie Containers and Lids
Buy Bain Marie Containers and Lids Items Including:
Bain Marie Container, HDPE, 12 quart
Bain Marie Container, HDPE, 16 quart
Bain Marie Container, HDPE, 20 quart
Bain Marie Container, HDPE, 24 quart
Bain Marie Container, HDPE, 32 quart
More Details
Thermo Scientific Orion Reference Fill Solutions
Buy Thermo Scientific Orion Reference Fill Solutions Items Including:
Thermo Scientific Orion electrode fill solution, 3 M KCl, 5 x 60mL bot
Optimum ResultsTM A filling solution: use with Ca+2/s (900061)
Optimum ResultsTM F filling solution: for use with NO3 (900046)
Filling solution f/ammonia ISE electrode (951202).
More Details
Thermo Scientific Orion Electrodes
Buy Thermo Scientific Orion Electrodes Items Including:
Thermo Scientific Orion electrode fill solution, 3 M KCl, 5 x 60mL bot
Thermo Scientific Orion pH electrode cleaning solution A, 4 x 30 mL bo
Thermo Scientific Orion pH electrode cleaning solution B, 4 x 60 mL bo.
More Details
Square Stainless Steel Mesh Sieve Pan
Buy Square Stainless Steel Mesh Sieve Pan Items Including:
Receiving Pan for 12" Stainless Steel Bottom Sieves, Full Height
Receiving Pan for 12" Stainless Steel Bottom Sieves, Half Height
Standard Sieve, Stainless Steel Mesh, 5.66 mm sieve opening, No
3.5.
More Details
Neotex Liners, Neoprene-Coated Fiber
Buy Neotex Liners, Neoprene-Coated Fiber Items Including:
NEOTEX neoprene-coated fiber liner, black
NEOTEX neoprene-coated fiber liner, red
NEOTEX neoprene-coated fiber liner, green.
More Details
Pyrex Brand 2103 Class a Burettes
Buy Pyrex Brand 2103 Class a Burettes Items Including:
PYREX Brand 2103 Burette; capacity, 10 mL; graduation interval, 0.05mL
PYREX Brand 2103 Burette; capacity, 25 mL; graduation interval, 0.10mL
PYREX Brand 2103 Burette; capacity, 50 mL; graduation interval, 0.10mL.
More Details
Micrometer Burettes and Syringes
Buy Micrometer Burettes and Syringes Items Including:
Gilmont micrometer burette, 2 mL
Gilmont micrometer burette tip, 2 mL, for use with 07846-00
Gilmont Micrometer Syringe, 0.2 mL
Gilmont micrometer syringe tip, for use with 07840-00
Gilmont micrometer syringe, 2 mL.
More Details
Digital Pressure Gauge with Output
Buy Digital Pressure Gauge with Output Items Including:
Ashcroft Digital Pressure Gauge, 0 to 1000 psi, 1/4" NPT(M) Process co (D1005PS2)
Additel Digital Pressure Gauge, 0" to 30" Hg, 1/4" NPT(M) Connection
Additel Digital Pressure Gauge, 0 to 100 psi, 1/4" NPT(M) Connection.
More Details
Pyrex Glass Graduated Cylinders
Buy Pyrex Glass Graduated Cylinders Items Including:
Pyrex Brand 3024 cylinder; 100 mL, pack of 1
Pyrex Brand 3024 cylinder; 250 mL, pack of 1
Pyrex Brand 3024 cylinder; 500 mL, pack of 1
Pyrex Brand 3024 cylinder; 10 mL, case of 24
Pyrex Brand 3024 cylinder; 25 mL, case of 18.
More Details
Gates Adapta Flex General Purpose Hose
Buy Gates Adapta Flex General Purpose Hose Items Including:
Gates Adapta Flex™ General Purpose Hose; ¼" x 0.47", 100 ft/pk
Gates Adapta Flex™ General Purpose Hose; 3/8" x 0.66", 100 ft/pk
Gates Adapta Flex™ General Purpose Hose; 5/8" x 0.93", 50 ft/pk.
More Details
10 Ml Glass Bottle Top Dispenser
Buy 10 Ml Glass Bottle Top Dispenser Items Including:
Cole-Parmer Bottletop Dispenser .25-2.5 mL with 100 mL Glass Bottle
Cole-Parmer Bottletop Dispenser 1-10 mL
Cole-Parmer Bottletop Dispenser 5-60 mL with 1000 mL Plastic Bottle
Cole-Parmer Bottletop Dispenser 2.5-30 mL with 500 mL Plastic Bottle.
More Details
Cotton Inspection Gloves
Buy Cotton Inspection Gloves Items Including:
100% Cotton Inspection Gloves, women's, 8"L, 12 pairs/pk
100% Cotton inspection gloves, men's, 9"L, 12 pairs/pk
Inspection Glove, Cotton, Mediumweight, Mens, 12 pairs/pk.
More Details
Rotary Vane Pump High Pressure
Buy Rotary Vane Pump High Pressure Items Including:
High-pressure, brass-bodied rotary vane pump, 0.75 GPM, 115/230 VAC
High-pressure, brass-bodied rotary vane pump, 2.3 GPM, 115/230 VAC, 5
High-pressure, 303 stainless steel rotary vane pump, 2.3 GPM, 115/230.
More Details
Kipp Group Male Luer Lock Fittings
Buy Kipp Group Male Luer Lock Fittings Items Including:
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
Luer stopcock, 1-way, male luer lock x male luer lock, black PVDF, pac (YO-31200-90)
One-way stopcocks; male luer lock x male luer lock, nickel-plated bras (YO-31507-02).
More Details
Kynar Male Luer Lock Fittings
Buy Kynar Male Luer Lock Fittings Items Including:
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
Luer stopcock, 1-way, male luer lock x male luer lock, black PVDF, pac (YO-31200-90)
One-way stopcocks; male luer lock x male luer lock, nickel-plated bras (YO-31507-02).
More Details
One-Way Luer Check Valves
Buy One-Way Luer Check Valves Items Including:
One-way luer check valve, SAN/LDPE with silicone diaphragm, female-mal (SCV08053).
More Details
Check Valve with Screen
Buy Check Valve with Screen Items Including:
PP Check Valve with built-in screen, 3/8" hose barb (YO-07091-55)
PP Check Valve with built-in screen, 1/2" hose barb (YO-07091-60)
PP Foot Valve with built-in screen, 3/8" hose barb (YO-07091-65).
More Details
Thermolyne Hot Plate
Buy Thermolyne Hot Plate Items Including:
Thermo Scientific Thermolyne Cast-Aluminum Hot Plate, 12" x 12", 120 V
Thermo Scientific Thermolyne Cast-Aluminum Hot Plate, 12" x 12", 240 V
Thermo Scientific Thermolyne Cast-Aluminum Hot Plate, 24" x 12", 120 V.
More Details
Kar Products Brass Fittings 95378
Buy Kar Products Brass Fittings 95378 Items Including:
Luer fittings, Nickel-plated brass fitting, male slip x 1/4" barb (YO-31507-60)
Nickel-plated brass fittings, male luer lock x 1/16" hose barb (YO-31507-62)
Nickel-plated brass fittings, female luer x 1/16" hose barb (YO-31507-64).
More Details
Tirrill Burner
Buy Tirrill Burner Items Including:
Bunsen burner, Tirrill burner, natural gas (H-6220).
More Details
70618
results
<<<Previous
12
13
14
15
16
17
18
19
20
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Whatman Filter Paper Sizes to Tirrill Burner
>
31125329-s-toc_2 31125329stoc2