Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 17
Alcatel 300 Vacuum Pump Oil Gallon
to
Mortar and Pestle Set
Image
Product Detail
More Detail
Alcatel 300 Vacuum Pump Oil Gallon
Buy Alcatel 300 Vacuum Pump Oil Gallon Items Including:
Cole-Parmer pump oil, 56 cSt, 1 gallon.
More Details
10 Ml Glass Centrifuge Tubes
Buy 10 Ml Glass Centrifuge Tubes Items Including:
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 10 ml, 12 (99502-10)
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 5 ml, 125 (99502-5)
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 15 ml, 12 (99502-15).
More Details
Wika Digital Pressure Gauges and Meters
Buy Wika Digital Pressure Gauges and Meters Items Including:
WIKA Stainless Steel Digital Gauge ~ 30" Hg to 29 psi, 1/4" NPT(M)
WIKA Stainless Steel Digital Gauge ~ 30" Hg to 72 psi, 1/4" NPT(M)
WIKA Stainless Steel Digital Gauge ~ 30" Hg to 145 psi, 1/4" NPT(M).
More Details
Encon Veratti 1000 Protective Eyewear
Buy Encon Veratti 1000 Protective Eyewear Items Including:
Encon Veratti 1000 Protective Eyewear, Clear Lens, Silver Frame.
More Details
Nalgene Sterile Petg Media Bottles
Buy Nalgene Sterile Petg Media Bottles Items Including:
Thermo Scientific Nalgene square polycarbonate bottle, 125 mL
Thermo Scientific Nalgene square polycarbonate bottle, 250 mL
Thermo Scientific Nalgene square polycarbonate bottle, 500 mL
Thermo Scientific Nalgene square polycarbonate bottle, 1 L.
More Details
Flat Surface Ph Electrode
Buy Flat Surface Ph Electrode Items Including:
Flat surface pH electrode (YO-98765-10)
Cole-Parmer self-cleaning pH electrode, PPS housing, 100 Ω Pt RT (YO-27003-23)
Cole-Parmer self-cleaning pH electrode, CPVC housing, 1 kΩ RTD,
Cole-Parmer self-cleaning pH electrode, CPVC housing, 100 kΩ the.
More Details
Stir Bar for 5 Gallon Bucket
Buy Stir Bar for 5 Gallon Bucket Items Including:
Scienceware Spinfin Stirring Bar, PTFE, 5/16" X 3/8"
Scienceware Spinfin Stirring Bar, PTFE, 5/8" X 1/2"
1" X 5/16" Disposable Stir Bars (80037-1066)
Large-Volume Stir Bar, 4-1/2"L x 1-1/16"dia (YO-04627-00).
More Details
Petri Dishes
Buy Petri Dishes Items Including:
Chemware Petri Dish PFA, 100 mL
Thermo Scientific Nunc™ Lab-Tek Sterile Petri Dishes, PS, Standard, (YO-14007-01)
Thermo Scientific Nunc™ Lab-Tek Sterile Petri Dishes, PS, Deep, 100
Sterile Petri Dishes; 100 mm dia x 15 mm H; 500/box (YO-06139-00).
More Details
Ionic Strength/Ph Adjusters
Buy Ionic Strength/Ph Adjusters Items Including:
Ionic strength and pH adjustor; Interference suppressor (930710)
Ionic strength and pH adjustor; SAOB reagent for S-2 (941609)
Ionic strength and pH adjustors, TISAB II (940909)
Ammonium sulfate adjuster 2m (930711)
Ion strength adjustor (951211).
More Details
Nalgene Lab Wash Bottles
Buy Nalgene Lab Wash Bottles Items Including:
Thermo Scientific Nalgene wide-mouth Unitary wash bottle, 1 L
Thermo Scientific Nalgene "Right-to-Know" safety wash bottle for ethan (2425-0502)
Thermo Scientific Nalgene "Right-to-Know" safety wash bottle for ethan (2425-1002).
More Details
Disposable Neoprene Gloves
Buy Disposable Neoprene Gloves Items Including:
Ansell NeoTouch Neoprene Gloves, small, 100/box
Ansell NeoTouch Neoprene Gloves, medium, 100/box
Ansell NeoTouch Neoprene Gloves, extra large, 100/box
MAPA Trilites Gloves, small
MAPA Trilites Gloves, medium
More Details
Stainless Barbed Elbow Fittings
Buy Stainless Barbed Elbow Fittings Items Including:
Elbow, Stainless Steel, 1/8", Each (YO-30620-13)
Elbow, Stainless Steel, 3/16", Each (YO-30620-14)
Elbow, Stainless Steel, 1/4", Each (YO-30620-15)
Barbed Fittings; NPT Male Pipe Adapter Elbow, 316 SS, 10-32 NPT(M) x 1 (YO-31209-45).
More Details
Hose Barb Reducing Connectors
Buy Hose Barb Reducing Connectors Items Including:
Barbed fittings, Reducing Connector, Natural PP,5/16" x 1/4" ID, 1/8", (YO-30610-07)
Barbed fittings, Reducing Connector, Natural PP, 3/8" x 1/4" ID, 1/8", (YO-30610-08).
More Details
Plastic Erlenmeyer Flask
Buy Plastic Erlenmeyer Flask Items Including:
Thermo Scientific Nalgene polypropylene Erlenmeyer flask, 250 mL
Thermo Scientific Nalgene polypropylene Erlenmeyer flask, 500 mL
Chemware Erlenmeyer Flask, PFA, 100 mL
Chemware Erlenmeyer Flask, PFA, 200 mL
Chemware Erlenmeyer Flask, PFA, 300 mL.
More Details
Cpvc Ball Valves
Buy Cpvc Ball Valves Items Including:
Thermoplastic True Union Ball Valve, CPVC with Viton seals, PTFE seats (V07104N)
Hayward electrically Actuated Ball Valve, 3-way, 1/2" CPVC (HCTN2050STV/EAU-28)
Hayward electrically Actuated Ball Valve, 3-way, 1" CPVC (HCTN2100STV/EAU-28).
More Details
Industrial Bench Scales
Buy Industrial Bench Scales Items Including:
Ohaus Trooper™ Compact Industrial Bench Scale, 120 VAC, 15 kg
Ohaus Trooper™ Compact Industrial Bench Scale, 240 VAC, 15 kg
Ohaus Trooper™ Compact Industrial Bench Scale, 120 VAC, 30 kg
Ohaus Trooper™ Compact Industrial Bench Scale, 240 VAC, 30 kg.
More Details
Normally Open Solenoid Valve
Buy Normally Open Solenoid Valve Items Including:
Two-way normally open solenoid pinch valve; 12 VDC, 1/32" ID x 3/32" O (075P2NO-12-01)
Two-way normally open solenoid pinch valve; 12 VDC, 1/16" ID x 1/8" OD (075P2NO-12-02).
More Details
Luer Stopcock
Buy Luer Stopcock Items Including:
Luer stopcock tubing assembly (YO-30526-04)
Stopcocks with Luer Connections; 1-way; male lock (YO-30600-00)
Stopcocks with Luer Connections; 1-way; male slip (YO-30600-01)
Stopcocks with Luer Connections; 1-way; male slip; 100/pack (YO-30600-06).
More Details
Bronze Gate Valves
Buy Bronze Gate Valves Items Including:
Bronze gate valve; 1" NPT(F) (GV0700150)
Bronze gate valve; 1-1/4" NPT(F) (GV0700160).
More Details
Ph Test Paper and Indicator Strips
Buy Ph Test Paper and Indicator Strips Items Including:
Universal pH Test Paper; pH Range 1 to 11; 9 mm x 6 m Roll
Litmus Red Test Paper, 10 Booklets of 20 Strips
pHydrion plastic pH indicator strips, 0.0 to 14.0, flip top vial packa
pHydrion plastic pH indicator strips, 0.0 to 14.0, clear plastic case.
More Details
British Standard Pipe Thread Fittings
Buy British Standard Pipe Thread Fittings Items Including:
Threaded adapter, stainless steel, 1/8" female NPT to 1/8" male BSPP (YO-31830-50).
More Details
Digital Temperature Chart Recorder
Buy Digital Temperature Chart Recorder Items Including:
Honeywell DR4300 10" Process Recorder with digital display, 2 channels
Honeywell DR4500A 12" Digital Recorder, 1 channel
Honeywell DR4500A 12" Digital Recorder, 2 channels
Honeywell DR4500A 12" Digital Recorder, 3 channels.
More Details
Low Temperature Chart Recorder
Buy Low Temperature Chart Recorder Items Including:
6" Chart Recorder for 3 cuft Revco CxF Ultra-Low Temperature Chest Fre
6" Ink Chart Recorder, Factory Installed
6" Inkless Chart Recorder, Factory Installed
6" Inkless Chart Recorder, Factory Installed, for UxF400, UxF500, UxF6.
More Details
Omega Temperature Chart Recorder
Buy Omega Temperature Chart Recorder Items Including:
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C304)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C312)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C320).
More Details
Refrigerator Temperature Chart Recorder
Buy Refrigerator Temperature Chart Recorder Items Including:
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C304)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C312)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C320).
More Details
Refrigerator Temperature Chart Recorder Laboratory
Buy Refrigerator Temperature Chart Recorder Laboratory Items Including:
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C304)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C312)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C320).
More Details
Temperature Chart Recorder Laboratory
Buy Temperature Chart Recorder Laboratory Items Including:
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C304)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C312)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C320).
More Details
Temperature Chart Recorder Manual
Buy Temperature Chart Recorder Manual Items Including:
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C304)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C312)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C320).
More Details
Temperature Chart Recorder Rent
Buy Temperature Chart Recorder Rent Items Including:
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C304)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C312)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C320).
More Details
Mortar and Pestle Set
Buy Mortar and Pestle Set Items Including:
Cole-Parmer Mortar and Pestle Set, Porcelain; 125ML, Each
Glass Mortar & Pestle Set, 100ml (YO-34500-53)
Glass Mortar & Pestle Set, 500ml 6/pk
Porcelain Pestle Grinding Set (YO-04181-51)
Sintered Corundum Pestle Grinding Set (YO-04181-53).
More Details
70618
results
<<<Previous
13
14
15
16
17
18
19
20
21
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Alcatel 300 Vacuum Pump Oil Gallon to Mortar and Pestle Set
>
31125329-s-toc_2 31125329stoc2