Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 18
Cole-Parmer Balance Draft Shield
to
Pure Gum Rubber Tubing
Image
Product Detail
More Detail
Cole-Parmer Balance Draft Shield
Buy Cole-Parmer Balance Draft Shield Items Including:
Cole-Parmer Balance Draft Shield, Small 13"W x 13-1/2"H x 14-3/4"D (small)
Cole-Parmer Balance Draft Sheild, Medium 15”W x 15-1/2”H x 16-3/4”D.
More Details
Plastic Fittings
Buy Plastic Fittings Items Including:
Bondable Fittings Kit, Polysulfone (YO-45520-10)
Quick-disconnect fittings, Polycarbonate fittings, PP, 1/4" ID, comp (YO-31305-00)
Quick-disconnect fittings, Polycarbonate fittings, PP, 1/4" ID, body, (MPC17004T03).
More Details
1 Cm Stir Bar
Buy 1 Cm Stir Bar Items Including:
Large-Volume Stir Bar, 4-1/2"L x 1-1/16"dia (YO-04627-00)
Large-Volume Stir Bar, 6-1/4"L x 1-1/16"dia (YO-04627-10)
1/2" X 1/8" Disposable Stir Bars (80037-1065)
Double Stir Bar, 1/2"H x 1/2"dia (37126-9002).
More Details
3 Way Ball Valve John Guest
Buy 3 Way Ball Valve John Guest Items Including:
Ball valve, 3-way, 1/4" John Guest (PVC-326-4SMC4SMC4SMC-E)
Ball valve, 2-way right angled, 3/8" NPT(F) x 3/8" John Guest (PVC-657-6F6CJG-B)
Ball valve, 2-way right angled, 3/8" NPT(F) x 3/8" John Guest (PVC-657-6F6CJG-E).
More Details
Ph Conductivity Meter
Buy Ph Conductivity Meter Items Including:
Thermo Scientific Orion 4-Star pH/Conductivity Meter Only
Thermo Scientific Orion 4-Star pH/Conductivity Meter Kit
accumet® XL20 pH/Conductivity Meter
Hach ISFET Benchtop pH/Conductivity Meter Only
Hach Waterproof ISFET pH/Conductivity Meter Only.
More Details
Stainless Steel Hose Clamps
Buy Stainless Steel Hose Clamps Items Including:
Stainless Steel (SS) Hose Clamps, 11/16 x 1-1/4", box of 10
Stainless Steel (SS) Hose Clamps, 13/16 x 1-3/4", box of 10
Stainless Steel (SS) Hose Clamps, 1-5/16 x 2-1/4", box of 10.
More Details
Walchem High Stroke-Frequency Diaphragm Metering Pumps
Buy Walchem High Stroke-Frequency Diaphragm Metering Pumps Items Including:
Remote Control Pump with PVC head, 1.3 GPH, 230 VAC, 50 Hz
Remote Control Pump with GFRPP head, 2.3 GPH, 115 VAC, 50/60 Hz
Remote Control Pump with GFRPP head, 2.3 GPH, 230 VAC, 50 Hz
Remote Control Pump with GFRPP head, 4.3 GPH, 115 VAC, 50/60 Hz.
More Details
Water Solenoid Valve
Buy Water Solenoid Valve Items Including:
Parker Two-way Solenoid Valve, 1/2 in NPT(F), 110/120 VAC, Steam and H
Parker Two-way Solenoid Valve, 3/4 in NPT(F), 110/120 VAC, Steam and H
Optional double solenoid valve for Thermo Scientific Barnstead Mega-Pu (440236).
More Details
Orion Reference Electrode Filling Solution
Buy Orion Reference Electrode Filling Solution Items Including:
ROSS electrode fill solution, 3 M KCl
Case of five 60-mL bottles (810007)
Thermo Scientific Orion electrode fill solution, 3 M KCl, 5 x 60mL bot
Filling solution sodium 5/cs (900012)
Optimum ResultsTM F filling solution: for use with NO3 (900046).
More Details
Cole-Parmer Ptfe Npt(M) Threaded Plugs
Buy Cole-Parmer Ptfe Npt(M) Threaded Plugs Items Including:
Threaded Plug, PTFE, 1/8" NPT(M) (F-PL01)
Threaded Plug, PTFE, 1/4" NPT(M) (F-PL02)
Threaded Plug, PTFE, 3/8" NPT(M) (F-PL03)
Threaded Plug, PTFE, 1/2" NPT(M) (F-PL04)
Threaded Plug, PTFE, 3/4" NPT(M) (F-PL06).
More Details
Filter Paper Whatman 1 150mm Dia
Buy Filter Paper Whatman 1 150mm Dia Items Including:
Whatman Qualitative Filter Papers; 5.5 cm dia; pore size, 20-25µ; 1
Whatman Qualitative Filter Papers; 15 cm dia; pore size, 20-25 µ; 1
Whatman Qualitative Filter Papers; 24 cm dia; pore size, 20-25 µ; 1.
More Details
Stainless Steel Wire Test Tube Racks
Buy Stainless Steel Wire Test Tube Racks Items Including:
Poxygrid Wire Test Tube Rack 72 10-13mm (F18750-0000)
Poxygrid Wire Test Tube Rack 120 10-13mm (YO-06739-16)
Poxygrid Wire Test Tube Rack 180 15-16mm (YO-06739-18)
Poxygrid Wire Test Tube Rack 60 10-13mm
Poxygrid Wire Test Tube Rack, 150, 10-13mm.
More Details
Nalgene Wash Bottles
Buy Nalgene Wash Bottles Items Including:
Thermo Scientific Nalgene Wash bottle, FEP, 1000mL, 1/pk
Thermo Scientific Nalgene "Right-to-Know" safety wash bottle for ethan (2425-0502)
Thermo Scientific Nalgene "Right-to-Know" safety wash bottle for ethan (2425-1002).
More Details
Conductivity Calibration Standards
Buy Conductivity Calibration Standards Items Including:
Conductivity calibration standard, 1000 µmhos (3161)
MYRON L conductivity calibration standard, 23.8 µS
MYRON L conductivity calibration standard, 46.7 µS
MYRON L conductivity calibration standard, 2060 µS.
More Details
Weiss Digital Solar-Powered Industrial Pressure Gauge
Buy Weiss Digital Solar-Powered Industrial Pressure Gauge Items Including:
Pressure Gauge Vacuum-15 PSI, 1/4" NPT(M); 316 SS Sensor/ 316 SS Socke
Pressure Gauge Vacuum-60 PSI, 1/4" NPT(M); 316 SS Sensor/ 316 SS Socke
Pressure Gauge Vacuum-100 PSI, 1/4" NPT(M); 316 SS Sensor/ 316 SS Sock.
More Details
Rite in the Rain
Buy Rite in the Rain Items Including:
Rite in the Rain - all weather copier paper - 8-1/2" x 11"
Rite in the Rain Field Pattern Maxi-Spiral Notebooks
Rite in the Rain Horizontal Line Pattern Maxi-Spiral Notebooks
4-5/8" x 7" spiral notebook - field pattern (353N-6).
More Details
Metal Luer Fitting Metric Thread
Buy Metal Luer Fitting Metric Thread Items Including:
Luer fittings, Chrome-plated Luer adapters,female luer to male luer lo (YO-31507-51)
Luer fittings, Chrome-plated Luer adapters,female luer plug (YO-31507-53)
Luer fittings, Chrome-plated Luer adapters,male luer lock to 1/8" to 3 (YO-31507-61).
More Details
Ysi do 200 Dissolved Oxygen Meters
Buy Ysi do 200 Dissolved Oxygen Meters Items Including:
YSI DO 200 Dissolved Oxygen Meter Only
YSI DO 200 Dissolved Oxygen Meter Kit, 13-ft (4-m) probe cables
YSI DO 200 Dissolved Oxygen Meter Kit, 33-ft (10-m) probe cables
EcoSense DO 200 Meter Only.
More Details
Stainless Steel Tubing Compression Fitting
Buy Stainless Steel Tubing Compression Fitting Items Including:
Compression To Thread Adapter, Stainless Steel, 6mm OD Tubing X 1/4" N (YO-31528-24)
Compression Straight Union, Stainless Steel, 10mm OD Tubing (YO-31528-35)
Compression Straight Union, Stainless Steel, 12mm OD Tubing (YO-31528-36).
More Details
Bca Protein Assay Kit
Buy Bca Protein Assay Kit Items Including:
Micro BCA™ Protein Assay Kit (YO-36490-02).
More Details
Blood Bank Supplies Test Tube Racks
Buy Blood Bank Supplies Test Tube Racks Items Including:
Scienceware test tube rack for 10- to 13-mm tubes
Scienceware polypropylene test tube rack for 26-30 mm tubes
Thermo Scientific Nalgene Unwire™ test tube rack for 13-mm tubes, w
Thermo Scientific Nalgene Unwire™ test tube rack for 13-mm tubes, o.
More Details
Safeskin Nitrile Gloves
Buy Safeskin Nitrile Gloves Items Including:
Kimberly-Clark Nitrile Gloves, 9.5" length, Medium; 100/box
Kimberly-Clark Nitrile Gloves, 9.5" length, Extra-large; 90/box
Kimberly-Clark Nitrile gloves, 12" length, Extra-large; 50/box
Kimberly-Clark Blue Nitrile Gloves, Medium.
More Details
Hepa Filtered Vacuum
Buy Hepa Filtered Vacuum Items Including:
Replacement HEPA Filter for Nilfisk Backpack Vacuum
Replacement HEPA filter
Replacement Exhaust Filter for Nilfisk Backpack HEPA Vacuum, pack of 3.
More Details
Female Luer Fitting
Buy Female Luer Fitting Items Including:
Adapters with caps; female luer fitting; Tefzel
Cole-Parmer Animal Free Female Luer Fittings, 3/16", PP, 25/pk
Cole-Parmer Animal Free Female Luer Fittings, PP, 25/pk
3-way "Y" adapter, 1 female luer, 2 male luer lock (YO-31507-81).
More Details
Latex Free Finger Cots
Buy Latex Free Finger Cots Items Including:
QRP™ Latex Antistatic Finger Cots, small
QRP™ Latex Antistatic Finger Cots, medium
QRP™ Latex Antistatic Finger Cots, extra-large
North Powdered Latex finger cots, small, 1440/bag
North Powdered Latex finger cots, medium, 1440/bag.
More Details
Portable Eyewash with Drench Hose
Buy Portable Eyewash with Drench Hose Items Including:
Portable Pressurized Eyewash with Drench Hose, 10 gallon
Portable Pressurized Eyewash with Drench Hose, 15 gallon
Wall-Mount Eyewash/Drench Hose Unit with Squeeze-Open Valve (G5026)
Wall-Mount Eyewash/Drench Hose Unit with Stay-Open Valve (G5014).
More Details
Push-Button Temperature Transmitters (Rtd, Thermocouple, Thermistor)
Buy Push-Button Temperature Transmitters (Rtd, Thermocouple, Thermistor) Items Including:
Economical push-button temperature transmitter, RTD (SEM203P)
Economical push-button temperature transmitter, thermocouple (SEM203).
More Details
Accessories for Heat Sealer
Buy Accessories for Heat Sealer Items Including:
Replacement heating elements for heat sealer, 8", 115 VAC (K200H)
Replacement heating elements for heat sealer, 8", 230 VAC (K205H)
Replacement heating elements for heat sealer, 12", 115 VAC (K300H).
More Details
Gum Rubber Tubing
Buy Gum Rubber Tubing Items Including:
Gum Rubber Tubing, 3/32" x 5/32", 25' per pack (057PXA-25').
More Details
Pure Gum Rubber Tubing
Buy Pure Gum Rubber Tubing Items Including:
Gum Rubber Tubing, 3/32" x 5/32", 25' per pack (057PXA-25')
Gum Rubber Tubing, 3/16" x 5/16", 25' per pack (099PXA-25')
Gum Rubber Tubing, 5/16" x 7/16", 25' per pack (138PXA-25').
More Details
70618
results
<<<Previous
14
15
16
17
18
19
20
21
22
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Cole-Parmer Balance Draft Shield to Pure Gum Rubber Tubing
>
31125329-s-toc_2 31125329stoc2