Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 19
Hayward True Union Solenoid Valves
to
Marsh Bellofram Type 1000 I
Image
Product Detail
More Detail
Hayward True Union Solenoid Valves
Buy Hayward True Union Solenoid Valves Items Including:
Hayward PVC True Union Solenoid Valve, 1/2", 110 VAC
Hayward CPVC True Union Solenoid Valve, 1/4", 110 VAC
Hayward CPVC True Union Solenoid Valve, 1/2", 110 VAC
Hayward CPVC True Union Solenoid Valve, 3/4", 110 VAC.
More Details
Pyrex Brand 3025 Cylinders
Buy Pyrex Brand 3025 Cylinders Items Including:
Cylinder, Pyrex Brand 3025, graduated, 10 mL
Cylinder, Pyrex Brand 3025, graduated, 100 mL
Cylinder, Pyrex Brand 3025, graduated, 250 mL
Cylinder, Pyrex Brand 3025, graduated, 500 mL
Cylinder, Pyrex Brand 3025, graduated, 100 mL, case of 12.
More Details
Polystyrene Foam Test Tube Racks
Buy Polystyrene Foam Test Tube Racks Items Including:
Foam Test Tube Rack-small, 6 per pack
Scienceware polypropylene test tube rack
Scienceware test tube rack for 10- to 13-mm tubes
Scienceware test tube rack for 13- to 16-mm tubes
Thermo Scientific Nalgene Unwire™ test tube rack for 13-mm tubes, w.
More Details
Ball Bearing Check Valve
Buy Ball Bearing Check Valve Items Including:
Hayward True Union Ball Check Valve, 1/2" socket and NPT(F) connectors
Hayward True Union Ball Check Valve, 3/4" socket and NPT(F) connectors
Hayward True Union Ball Check Valve, 1-1/2" socket and NPT(F) connecto.
More Details
Ball Cone Check Valves
Buy Ball Cone Check Valves Items Including:
Hayward True Union Ball Check Valve, 1/2" socket and NPT(F) connectors
Hayward True Union Ball Check Valve, 3/4" socket and NPT(F) connectors
Hayward True Union Ball Check Valve, 1-1/2" socket and NPT(F) connecto.
More Details
Check Valve Ball Retainer
Buy Check Valve Ball Retainer Items Including:
Hayward True Union Ball Check Valve, 1/2" socket and NPT(F) connectors
Hayward True Union Ball Check Valve, 3/4" socket and NPT(F) connectors
Hayward True Union Ball Check Valve, 1-1/2" socket and NPT(F) connecto.
More Details
Eslom Ball Check Valve
Buy Eslom Ball Check Valve Items Including:
Hayward True Union Ball Check Valve, 1/2" socket and NPT(F) connectors
Hayward True Union Ball Check Valve, 3/4" socket and NPT(F) connectors
Hayward True Union Ball Check Valve, 1-1/2" socket and NPT(F) connecto.
More Details
Inline Ball Check Valve Gasoline
Buy Inline Ball Check Valve Gasoline Items Including:
Hayward True Union Ball Check Valve, 1/2" socket and NPT(F) connectors
Hayward True Union Ball Check Valve, 3/4" socket and NPT(F) connectors
Hayward True Union Ball Check Valve, 1-1/2" socket and NPT(F) connecto.
More Details
Whitey 1 8 Needle Valves
Buy Whitey 1 8 Needle Valves Items Including:
Chemically Inert Panel-Mount Needle Valve, 1/2" NPT(M) (NV8-8-3)
Chemically Inert Panel-Mount Needle Valve, 1/8" NPT(M) (NV2-2-1)
Chemfluor® PTFE Needle Valve, 1/8" NPT(F) (D1071796)
PVDF Needle Valve, 3/8" NPT(F) (155.522.555).
More Details
Luer Lock Fittings Threaded
Buy Luer Lock Fittings Threaded Items Including:
Snap luer lock ring, PC, 25/pk (YO-45504-74)
Snap luer lock ring, Nylon, 25/pk (YO-45505-74)
Nickel-plated brass fittings, male luer lock x 10-32 UNF thread (YO-31507-70)
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55).
More Details
Ashcroft Pressure Transducer
Buy Ashcroft Pressure Transducer Items Including:
Differential Pressure Transducer, 0.25" Water Column, 4-20 mA output (CXLDP0.25IWC) (YO-68333-20)
Differential Pressure Transducer, 2.0" Water Column, 4-20 mA output
Differential Pressure Transducer, 5.0" Water Column, 4-20 mA output.
More Details
Scienceware Autoclavable Biohazard Disposable Bags, Hdpe
Buy Scienceware Autoclavable Biohazard Disposable Bags, Hdpe Items Including:
Scienceware Autoclavable HDPE Biohazard Disposal Bags with Sterilizati (YO-06497-30)
Scienceware Autoclavable HDPE Biohazard Disposal Bags with Sterilizati (YO-06497-40).
More Details
Laboratory Tube Racks
Buy Laboratory Tube Racks Items Including:
Thermo Scientific Nalgene Unwire™ test tube rack for 20-mm tubes, w
Thermo Scientific Nalgene Unwire™ test tube rack for 25-mm tubes, w
Poxygrid Wire Test Tube Rack 120 10-13mm (YO-06739-16)
Cole-Parmer Test Tube Rack, PP 24 places (YO-06739-17).
More Details
Vacuum Isolation Valve
Buy Vacuum Isolation Valve Items Including:
Solenoid vacuum valve; right-angle, NW16, aluminum, 110 VAC
Solenoid vacuum valve; right-angle, NW25, aluminum, 110 VAC
Solenoid vacuum valve; right-angle, NW25, aluminum, 240 VAC
Solenoid vacuum valve; in-line, NW16, aluminum, 110 VAC.
More Details
Thermo Scientific Nalgene Safety Products
Buy Thermo Scientific Nalgene Safety Products Items Including:
Thermo Scientific Nalgene Fluorinated HDPE Safety Waste System
Thermo Scientific Nalgene "Right-to-Know" safety wash bottle for ethan (2425-0502)
Thermo Scientific Nalgene "Right-to-Know" safety wash bottle for ethan (2425-1002).
More Details
Stainless Sanitary Fittings
Buy Stainless Sanitary Fittings Items Including:
Cole-Parmer, SANITARY CLAMP ELBOW, 90 degree, 316L SS, 3/4"
Cole-Parmer, Sanitary Clamp Elbow, 90 Degree, 316L SS, 1"
Cole-Parmer, Sanitary Clamp Reducer, 316L SS, 1" X 3/4"
Cole-Parmer, Sanitary Clamp Tee, 304L SS, 1-1/2".
More Details
Stainless Steel Trays
Buy Stainless Steel Trays Items Including:
Stainless Steel Utility Tray, 17-1/4" L x 2" W x 4-1/2" D
Stainless Steel Utility Tray, 8 7/8"L x 5"W x 2"D (952)
Stainless Steel Cover for tray 07276-06 (952D-2)
Stainless steel utility tray, 10"L x 6-1/2"W.
More Details
Gas Cylinder Safety Straps
Buy Gas Cylinder Safety Straps Items Including:
Safety Strap with secure Message
Safety Strap without Message
Heavy Duty Bench Clamp
Chain Assembly Wall/Bench
Replacement Wall Bracket
More Details
Stainless Steel Mesh Sieve Set
Buy Stainless Steel Mesh Sieve Set Items Including:
Receiving Pan for 12" Stainless Steel Bottom Sieves, Full Height
Receiving Pan for 12" Stainless Steel Bottom Sieves, Half Height
Standard Sieve, Stainless Steel Mesh, 5.66 mm sieve opening, No
3.5.
More Details
Flexible Stainless Steel Tubing
Buy Flexible Stainless Steel Tubing Items Including:
316 Stainless Steel Tubing; dimensions, 1/16" ID, 1/8" OD, 1/32" wall; (YO-03300-05)
304 Stainless Steel Tubing, 1/8" OD, 0.085" ID, 50ft roll (YO-34680-50)
Stainless steel tube set for electric and air motors, 27"L.
More Details
Polished Stainless Steel Tubing
Buy Polished Stainless Steel Tubing Items Including:
316 Stainless Steel Tubing; dimensions, 1/16" ID, 1/8" OD, 1/32" wall; (YO-03300-05)
304 Stainless Steel Tubing, 1/8" OD, 0.085" ID, 50ft roll (YO-34680-50)
Stainless steel tube set for electric and air motors, 27"L.
More Details
Small Stainless Steel Tubing
Buy Small Stainless Steel Tubing Items Including:
316 Stainless Steel Tubing; dimensions, 1/16" ID, 1/8" OD, 1/32" wall; (YO-03300-05)
304 Stainless Steel Tubing, 1/8" OD, 0.085" ID, 50ft roll (YO-34680-50)
Stainless steel tube set for electric and air motors, 27"L.
More Details
Cole-Parmer Stainless Steel Cell Spreaders
Buy Cole-Parmer Stainless Steel Cell Spreaders Items Including:
Cell spreader for 60-mm plates, 16-mm spreading bar, stainless steel (YO-14212-00).
More Details
Wooden Test Tube Rack
Buy Wooden Test Tube Rack Items Including:
Scienceware test tube rack for 10- to 13-mm tubes
Scienceware polypropylene test tube rack for 26-30 mm tubes
Thermo Scientific Nalgene Unwire™ test tube rack for 13-mm tubes, w
Thermo Scientific Nalgene Unwire™ test tube rack for 13-mm tubes, o.
More Details
Stainless Luer Lock Fittings
Buy Stainless Luer Lock Fittings Items Including:
316 SS fittings; female luer x male luer lock (YO-31507-21).
More Details
Steel Luer Lock Fittings
Buy Steel Luer Lock Fittings Items Including:
316 SS fittings; female luer x male luer lock (YO-31507-21).
More Details
Jars for Roller Mill
Buy Jars for Roller Mill Items Including:
One-tier jar high-capacity laboratory jar mill; 13" roller, 115 VAC/60 (YO-04149-00)
One-tier jar high-capacity laboratory jar mill; 13" roller, 230 VAC/50 (YO-04149-05).
More Details
Laboratory Roller Mill
Buy Laboratory Roller Mill Items Including:
One-tier jar high-capacity laboratory jar mill; 13" roller, 115 VAC/60 (YO-04149-00)
One-tier jar high-capacity laboratory jar mill; 13" roller, 230 VAC/50 (YO-04149-05).
More Details
Mini Roller Jar Mills
Buy Mini Roller Jar Mills Items Including:
One-tier jar high-capacity laboratory jar mill; 13" roller, 115 VAC/60 (YO-04149-00)
One-tier jar high-capacity laboratory jar mill; 13" roller, 230 VAC/50 (YO-04149-05).
More Details
Marsh Bellofram Type 1000 I/P Transducers
Buy Marsh Bellofram Type 1000 I/P Transducers Items Including:
Pressure Transducer, I to P;3-15 PSI Output, 4-20 mA Input (961-070-000)
Pressure Transducer, I to P;3-27 PSI Output, 4-20 mA Input (961-074-000)
Pressure Transducer, I to P;6-30 PSI Output, 4-20 mA Input (961-075-000).
More Details
70618
results
<<<Previous
15
16
17
18
19
20
21
22
23
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Hayward True Union Solenoid Valves to Marsh Bellofram Type 1000 I
>
31125329-s-toc_2 31125329stoc2