Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths & Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cleaning & Janitorial Supplies
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Lab Supplies & Consumables
Laboratory Equipment
Life Science
Masterflex® Peristaltic Pumps
Masterflex® Pump Tubing
Material Storage & Handling
Material Testing & Physical Testing
Microscopes & Magnification
Mixers and Blenders
pH, Conductivity, Dissolved Oxygen, and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity and Moisture Products
Tools
Tubing, Fittings, and Hose
Product Index Page 2
Oakton Thermocouple Probes
to
Cole-Parmer Chlorob
Image
Product Detail
More Detail
Oakton Thermocouple Probes
Buy Oakton Thermocouple Probes Items Including:
Type J All stainless steel thermocouple probe; 8"L (050797A-J)
Type K All stainless steel thermocouple probe; 8"L (050797A-K)
Type J High Temperature 12"L, .040" diameter thermocouple probe with s (073098D-J-G-12).
More Details
Cole-Parmer Chlorop to Chloroq
Buy Cole-Parmer Chlorop to Chloroq Items Including:
Chlorophenol Red Indicator, 0.04% (w/v) Aqueous Solution (500 mL) (YO-86970-95)
3-Chloropropionic acid 98% (100g) (YO-88164-11)
3-Chloropropionic acid 98% (500g) (YO-88164-12)
1-(4-Chlorophenyl)-1-cyclopentanecarboxylic acid 98% (5g) (YO-88169-57).
More Details
Peristaltic Metering Pumps
Buy Peristaltic Metering Pumps Items Including:
Pump, Cole-Parmer Deluxe Peristaltic Metering Pump, Max flow of 17.4 G (YO-74200-56)
Pump, Cole-Parmer Deluxe Peristaltic Metering Pump, Max flow of 33.3 G
Pump, Cole-Parmer Large Peristaltic Metering Pump, Max flow of 54 GPH, (YO-74203-64).
More Details
Cole-Parmer Diamino to Dibenzy
Buy Cole-Parmer Diamino to Dibenzy Items Including:
1,3-Diamino-2-hydroxypropane 95% (5g) (YO-88103-79)
D-Lactose Monohydrate (Powder/NF) (2.5kg) (YO-88075-54)
D-Lactose Monohydrate (Powder/NF) (500g) (YO-88075-57)
3,3'-Diaminobenzidine tetrahydrochloride,97%, contains up to 10% water (YO-88103-64).
More Details
Cole-Parmer Ca to Calycot
Buy Cole-Parmer Ca to Calycot Items Including:
Calcium Standard, 1 mL = 10 mg Ca, 10,000 ppm Ca (500 mL) (YO-86970-79)
Cadmium AA Std, 1000 ppm Cd, Metal in Nitric, 100mL (YO-86995-24)
Cadmium AA Std, 1000 ppm Cd, Metal in Nitric, 500mL (YO-86995-25)
Cadmium ICP 1000 ppm 100ML (YO-86995-26).
More Details
Cole-Parmer Dimethyl to Dimethyla
Buy Cole-Parmer Dimethyl to Dimethyla Items Including:
N,N-Dimethylacetamide 99% (2.5l) (YO-88109-42)
N,N-Dimethylacetamide 99% (5ml) (YO-88109-43)
4-Dimethylaminoantipyrine 97% (5g) (YO-88109-49)
3-Dimethylaminobenzoic acid 99% (25g) (YO-88109-51)
3-Dimethylaminobenzoic acid 99% (100g) (YO-88109-52).
More Details
Cole-Parmer Valves
Buy Cole-Parmer Valves Items Including:
Cole-Parmer Concentration Contoller-Air Solenoid Valve (mounted), 3/8& (YO-78120-03)
Cole-Parmer® Solenoid Pinch Valve; for use with L/S® 14 tubing (PK-1605NC-12V)
Cole-Parmer® Solenoid Pinch Valve; for use with L/S® 14 tubing (PK-1605NC-24V).
More Details
Cole-Parmer Methylp to Methylq
Buy Cole-Parmer Methylp to Methylq Items Including:
Methyl Purple Indicator Solution (500 mL) (YO-86973-59)
Methyl Purple Indicator Solution (32 oz) (YO-86980-73)
N-Methylpiperidine 99% (250ml) (YO-88128-12)
2-Methylpiperidine 99% (100ml) (YO-88128-14)
2-Methylpiperidine 99% (500ml) (YO-88128-15).
More Details
Cole-Parmer Trifluoro to Trimell
Buy Cole-Parmer Trifluoro to Trimell Items Including:
α,α,α-Trifluoro-m-cresol 98+% (25ml) (YO-88178-95)
α,α,α-Trifluoro-m-cresol 98+% (100ml) (YO-88178-96)
Trifluoroacetic Acid (Clear, Fuming Liq./Biochemical Grade) (100mL) (YO-88078-23)
Trifluoroacetaldehyde ethyl hemiacetal 90% (5ml) (YO-88142-11).
More Details
Solenoid Electric Valves (Actuated)
Buy Solenoid Electric Valves (Actuated) Items Including:
Two-Way, Pilot-Operated Valve with Manual Override, 1/2" NPT(F), 120 V (453349M)
Two-Way, Pilot-Operated Valve with Manual Override, 3/4" NPT(F), 120 V (453350J).
More Details
Cole-Parmer Methyli to Methylo
Buy Cole-Parmer Methyli to Methylo Items Including:
Methyl Orange Indicator, 0.1% (w/v) Aqueous Solution (500 mL) (YO-86973-56)
2-Methylimidazole 99% (500g) (YO-88127-38)
N-Methyliminodiacetic acid 99% (5g) (YO-88127-39)
3-Methylindole 98% (10g) (YO-88127-42)
3-Methylindole 98% (25g) (YO-88127-43).
More Details
Cole-Parmer Methylc to Methyld
Buy Cole-Parmer Methylc to Methyld Items Including:
Methyl cyclopropanecarboxylate 98% (100g) (YO-88126-91)
Methyl 2,4-dihydroxybenzoate 97% (25g) (YO-88126-95)
N-Methyl-D-glucamine 99% (100g) (YO-88127-11)
N-Methyl-D-glucamine 99% (500g) (YO-88127-12)
5-Methyl-DL-tryptophan 98% (1g) (YO-88128-83).
More Details
Cole-Parmer Safety Products
Buy Cole-Parmer Safety Products Items Including:
Cole-Parmer® Raised Diamond Grip Nitrile Gloves, single-layer, lig (YO-40216-02)
Cole-Parmer® Raised Diamond Grip Nitrile Gloves, single-layer, lig (YO-40216-03)
Cole-Parmer® Raised Diamond Grip Nitrile Gloves, single-layer, lig (YO-40216-04).
More Details
Cole-Parmer Da to Diallyl
Buy Cole-Parmer Da to Diallyl Items Including:
Da 40 Nmol CPg Column 10/pk (YO-88062-75)
2'-Deoxyadenosine-Long Chain Alkyl Amino (dA-LCAA-CPG)/Prepacked C (YO-88062-79)
Deoxycholic Acid Sodium Salt (100g) (YO-88066-69)
Dextrose Anhydrous (Cryst
Granules/Molecular Biology) (1kg) (YO-88066-70).
More Details
Cole-Parmer Dilaury to Dimetho
Buy Cole-Parmer Dilaury to Dimetho Items Including:
1,2-Dimethoxyethane (Certified) (1 liter) (YO-88077-27)
1,2-Dimethoxyethane (Certified) (4 liter) (YO-88077-28)
3,4-Dimethoxybenzoic acid 99+% (100g) (YO-88108-98)
3,4-Dimethoxybenzyl alcohol 96% (100g) (YO-88109-06).
More Details
Cole-Parmer Leb to Lyxosem
Buy Cole-Parmer Leb to Lyxosem Items Including:
Linear Alkylbenzene Sulfonate (LAS) Standard, 1 mL = 0.01 mg LAS, 10 p (YO-86973-14)
Leukotriene B4 (25ug) (YO-88060-45)
Leukotriene C4 (25ug) (YO-88060-46)
Leupeptin (Hemisulfate) (1mg) (YO-88061-76)
Leupeptin (Hemisulfate) (100mg) (YO-88061-77).
More Details
Cole-Parmer Barbed to Threaded Adapters
Buy Cole-Parmer Barbed to Threaded Adapters Items Including:
Threaded adapter elbows, nylon, 3/4" NPTM x 5/8" ID, 100/pack (YO-30401-26)
Threaded adapter, nylon, 1" NPTM x 1" tubing ID, 10/pack (YO-30401-34)
Threaded adapter, nylon, 1" NPTM x 1" tubing ID, 100/pack (YO-30401-35).
More Details
Lab Frames and Rods
Buy Lab Frames and Rods Items Including:
Aluminum Lab Frame, Small (916173)
Aluminum Lab Frame, Heavy-duty (916178)
Stainless Steel Lab Frame, Small (916197)
Stainless Steel Lab Frame, Medium (916198)
Stainless Steel Lab Frame, Large (916199)
More Details
Cole-Parmer Phenyls to Phospho
Buy Cole-Parmer Phenyls to Phospho Items Including:
Phenyl sulfide 99% (100ml) (YO-88133-44)
Phosphate Standard, 1 mL = 0.5 mg PO43-, 500 ppm (YO-86974-51)
Phosphoric Acid, 20% (v/v) Aqueous Solution (1 + 4) (500 mL) (YO-86974-55)
Phloroglucinol TS, 2% (w/v) Alcoholic Solution (32 oz) (YO-86981-20).
More Details
Cole-Parmer Compression Fittings
Buy Cole-Parmer Compression Fittings Items Including:
Compression fittings, Straight union, White PP, 1/4" OD, 1/4", 1", 5/ (YO-30802-94)
Compression fittings, Union Tees, Kynar, 1/8" OD, 1-1/8", 3/4", 3/32"; (YO-30805-21).
More Details
Cole-Parmer Ia to Iodine
Buy Cole-Parmer Ia to Iodine Items Including:
Iodine (Iodine-Iodide), 0.0100 Normal (N/100) (500 mL) (YO-86972-89)
Icp-ms Cal Std-1 100ml (YO-86996-95)
Icp-ms Cal Std-2 100ml (YO-86996-96)
Icp-ms Cal Std-100 100ml (YO-86996-98)
Infusorial Earth (White, Calcined Powder) (3kg) (YO-88027-14).
More Details
Form Liquid
Buy Form Liquid Items Including:
Low Form Dewar Flasks, 275mL (8120)
Low Form Dewar Flasks, 1000mL (8140)
Low Form Dewar Flasks, 2000mL (8150)
H-B Instruments Hydrometer, —1 to 11° API, Plain Form
DURAC SAFETYBLUE SPECIFIC GRAVITY COMBINED FORM HYDROMETER, 1.800 TO 2.
More Details
Industrial Pressure Gauges
Buy Industrial Pressure Gauges Items Including:
Cole-parmer Industrial Pressure Gauge, 30" Hg To 150 PSI; 1/4" NPT(M)
Cole-parmer Industrial Pressure Gauge, 30" Hg To 400 PSI; 1/4" NPT(M)
Cole-parmer Industrial Pressure Gauge, 30" Hg To 200 PSI; 1/4" NPT(M).
More Details
Cole-Parmer Ethylb to Ethyld
Buy Cole-Parmer Ethylb to Ethyld Items Including:
N-Ethylcarbazole 98.5% (100g) (YO-88113-52)
N-Ethylcarbazole 98.5% (500g) (YO-88113-53)
Ethyl chloroacetate 99% (2.5l) (YO-88113-55)
Ethyl 4-chlorobutyrate 97% (100g) (YO-88113-59)
Ethyl cyanoacetate 98+% (250g) (YO-88113-65).
More Details
Cole-Parmer Clamps and Supports
Buy Cole-Parmer Clamps and Supports Items Including:
Frame Clamps, T-Joint, Pack of 5 (53-2032W)
Frame Clamps, In-line, Pack of 5 (53-2042W)
Two-prong extension clamp, Dual, Large (916059)
Ss Standard Three-prong Extension Clamp/small (916352)
Ss Standard Three-prong Extension Clamp Medium (916351).
More Details
Cole-Parmer Bromof to Bromom
Buy Cole-Parmer Bromof to Bromom Items Including:
2-Bromofluorobenzene 99% (10ml) (YO-88095-48)
2-Bromofluorobenzene 99% (50ml) (YO-88095-49)
4-Bromo-2-methylaniline 97% (25g) (YO-88177-52)
5-Bromo-2-methoxybenzylalcohol 97% (5g) (YO-88196-65)
2-Bromo-6-methoxynaphthalene 98% (25g) (YO-88203-62).
More Details
Cole-Parmer Phenylg to Phenylq
Buy Cole-Parmer Phenylg to Phenylq Items Including:
DL-2-Phenylpropionic acid 98% (25g) (YO-88133-34)
N-Phenyl-1-naphthylamine 98% (25g) (YO-88145-94)
N-Phenyl-1-naphthylamine 98% (250g) (YO-88145-95)
L(-)-3-Phenyllactic acid 98% (1g) (YO-88152-74)
L(-)-3-Phenyllactic acid 98% (5g) (YO-88152-75).
More Details
Cole-Parmer Benzem to Benzoyl
Buy Cole-Parmer Benzem to Benzoyl Items Including:
Benzopinacole 98% (100g) (YO-88093-40)
1,4-Benzoquinone 98% (1kg) (YO-88093-42)
1,4-Benzoquinone 98% (500g) (YO-88093-44)
1,4-Benzoquinone dioxime, moistened with ca
30% water 94% (25g) (YO-88093-45)
More Details
Advantec Filter Papers
Buy Advantec Filter Papers Items Including:
Advantec Grade No
1 Qualitative Filter Paper; 5.5 cm dia.; pack of 10
Advantec Grade No
1 Qualitative Filter Paper; 7.0 cm dia.; pack of 10
Advantec Grade No
More Details
Cole-Parmer Chlorob to Chloroc
Buy Cole-Parmer Chlorob to Chloroc Items Including:
Chlorobenzene (Certified) (500mL) (YO-88005-16)
Chlorobenzene (Certified) (1 liter) (YO-88005-18)
4-Chlorobenzoyl chloride 99+% (500ml) (YO-88151-04)
4-Chlorobenzaldehyde 98.5+% (1kg) (YO-88151-65)
4-Chlorobenzaldehyde 98.5+% (100g) (YO-88151-66).
More Details
64361
results
<<<Previous
1
2
3
4
5
6
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,146
Oakton Thermocouple Probes to Cole-Parmer Chlorob to Chloroc
31125329-s-toc_2 31125329stoc2