Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 20
Thermo Scientific Thermolyne Atmosphere-Controlled Muffle Furnaces
to
1 Micron Nylon Filter Bags
Image
Product Detail
More Detail
Thermo Scientific Thermolyne Atmosphere-Controlled Muffle Furnaces
Buy Thermo Scientific Thermolyne Atmosphere-Controlled Muffle Furnaces Items Including:
Thermo Scientific Thermolyne Atmosphere-Controlled Programmable Muffle (F6020C-33-60-80).
More Details
Kimax Volumetric Flasks
Buy Kimax Volumetric Flasks Items Including:
Kimax Wide-Mouth Volumetric Flask, 5mL, PTFE stopper, 6/cs
Kimax Wide-Mouth Volumetric Flask, 10mL, PTFE stopper, 6/cs
KIMAX Square Volumetric Flask, 100mL 1/pk
KIMAX Square Volumetric Flask, 1000mL 1/pk
KIMAX Square Volumetric Flask, 2000mL 1/pk.
More Details
Ansell Hyflex Work Gloves
Buy Ansell Hyflex Work Gloves Items Including:
Ansell HyFlex™ Work Gloves, Size 7
Ansell HyFlex™ Lite Work Gloves, Size 7
Ansell HyFlex™ Lite Work Gloves, Size 10
Ansell® HyFlex®Cut Resistant Dyneema® Work Gloves, Size 10 (288738)
Ansell® HyFlex® Nylon Kit Work Gloves with Nitrile AGT, Size 7.
More Details
Ammex Black Nitrile Gloves
Buy Ammex Black Nitrile Gloves Items Including:
Best Nighthawk™ Black Nitrile Gloves, X-large, Box of 50
Microflex MidKnight™ Black Nitrile Glove, small, 100/box
Microflex MidKnight™ Black Nitrile Glove, medium, 100/box
Microflex MidKnight™ Black Nitrile Glove, extra large, 100/box.
More Details
Black Lightning Nitrile Gloves
Buy Black Lightning Nitrile Gloves Items Including:
Best Nighthawk™ Black Nitrile Gloves, X-large, Box of 50
Microflex MidKnight™ Black Nitrile Glove, small, 100/box
Microflex MidKnight™ Black Nitrile Glove, medium, 100/box
Microflex MidKnight™ Black Nitrile Glove, extra large, 100/box.
More Details
Black Magic Nitrile Gloves
Buy Black Magic Nitrile Gloves Items Including:
Best Nighthawk™ Black Nitrile Gloves, X-large, Box of 50
Microflex MidKnight™ Black Nitrile Glove, small, 100/box
Microflex MidKnight™ Black Nitrile Glove, medium, 100/box
Microflex MidKnight™ Black Nitrile Glove, extra large, 100/box.
More Details
Black Nitrile Gloves
Buy Black Nitrile Gloves Items Including:
Best Nighthawk™ Black Nitrile Gloves, X-large, Box of 50
Microflex MidKnight™ Black Nitrile Glove, small, 100/box
Microflex MidKnight™ Black Nitrile Glove, medium, 100/box
Microflex MidKnight™ Black Nitrile Glove, extra large, 100/box.
More Details
Shadow Black Nitrile Gloves
Buy Shadow Black Nitrile Gloves Items Including:
Best Nighthawk™ Black Nitrile Gloves, X-large, Box of 50
Microflex MidKnight™ Black Nitrile Glove, small, 100/box
Microflex MidKnight™ Black Nitrile Glove, medium, 100/box
Microflex MidKnight™ Black Nitrile Glove, extra large, 100/box.
More Details
Wire Test Tube Racks
Buy Wire Test Tube Racks Items Including:
Poxygrid Wire Test Tube Rack 72 10-13mm (F18750-0000)
Poxygrid Wire Test Tube Rack 120 10-13mm (YO-06739-16)
Poxygrid Wire Test Tube Rack 180 15-16mm (YO-06739-18)
Poxygrid Wire Test Tube Rack 60 10-13mm
Poxygrid Wire Test Tube Rack, 150, 10-13mm.
More Details
Industrial Cleaning Supplies
Buy Industrial Cleaning Supplies Items Including:
Industrial strength ultrasonic cleaning solution, 1 quart bottle (YO-08801-21)
Industrial strength ultrasonic cleaning solution, 5 gallon bottle (YO-08801-27).
More Details
2 Full Flow Check Valve
Buy 2 Full Flow Check Valve Items Including:
Hayward True Union Ball Check Valve, 2" socket and NPT(F) connectors
Brass Check Valve, 1/2" NPT(F) connectors (0376371)
PVC Check Valve, 1/2" NPT(F) connections (CKM050V-PV)
PP Check Valve, 1/2" NPT(F) connections (CKM050V-PP).
More Details
Infusion Tubing Luer Fittings
Buy Infusion Tubing Luer Fittings Items Including:
Plastic luer fittings, PVC tubing with luer ends, 48" L, M/F fitting e (YO-30600-61)
Luer fittings, Chrome-plated Luer adapters,female luer to male luer lo (YO-31507-51)
Luer fittings, 316 SS fittings, female luer x male slip (YO-31507-20).
More Details
Luer Fittings Peek Tubing
Buy Luer Fittings Peek Tubing Items Including:
Plastic luer fittings, PVC tubing with luer ends, 48" L, M/F fitting e (YO-30600-61)
Luer fittings, Chrome-plated Luer adapters,female luer to male luer lo (YO-31507-51)
Luer fittings, 316 SS fittings, female luer x male slip (YO-31507-20).
More Details
Electric Actuated Ball Valve Stainless Steel
Buy Electric Actuated Ball Valve Stainless Steel Items Including:
Hayward electrically Actuated Ball Valve, 3-way, 1" PVC (HCTN1100STV/EAU-28)
Hayward electrically Actuated Ball Valve, 3-way, 2" PVC (HCTN1200STV/EAU-28)
Hayward electrically Actuated Ball Valve, 3-way, 1/2" CPVC (HCTN2050STV/EAU-28).
More Details
Large Funnels
Buy Large Funnels Items Including:
Large industrial-size low-density polyethylene funnel, 1.5 L (YO-06121-10)
Large industrial-size polypropylene funnel, 12 L (YO-06121-00)
Large industrial-size polypropylene funnel, 4.5 L (YO-06121-32)
Weighing Funnels, Glass, Large (YO-34700-06).
More Details
Laboratory Frame/Rod/Clamp Holders
Buy Laboratory Frame/Rod/Clamp Holders Items Including:
Jumbo Clamp Holder, Stainless steel
Heavy-duty clamp (YO-04340-50)
Swivel Holder, Nickel-plated zinc
Regular Holder, Nickel-Plated
Regular Holder, Stainless Steel (110B)
More Details
Cole-Parmer Bottle Top Dispensers
Buy Cole-Parmer Bottle Top Dispensers Items Including:
Cole-Parmer Bottletop Dispenser .25-2.5 mL
Cole-Parmer Bottletop Dispenser .25-2.5 mL with 100 mL Glass Bottle
Cole-Parmer Bottletop Dispenser 2.5-30 mL with 500 mL Glass Bottle
Cole-Parmer Bottletop Dispenser 5-60 mL with 1000 mL Glass Bottle.
More Details
Tall Test Tube Racks
Buy Tall Test Tube Racks Items Including:
Scienceware test tube rack for 10- to 13-mm tubes
Scienceware polypropylene test tube rack for 26-30 mm tubes
Thermo Scientific Nalgene Unwire™ test tube rack for 13-mm tubes, w
Thermo Scientific Nalgene Unwire™ test tube rack for 13-mm tubes, o.
More Details
Test Tube Racks Metal
Buy Test Tube Racks Metal Items Including:
Scienceware test tube rack for 10- to 13-mm tubes
Scienceware polypropylene test tube rack for 26-30 mm tubes
Thermo Scientific Nalgene Unwire™ test tube rack for 13-mm tubes, w
Thermo Scientific Nalgene Unwire™ test tube rack for 13-mm tubes, o.
More Details
Test Tube Racks Nasco
Buy Test Tube Racks Nasco Items Including:
Scienceware test tube rack for 10- to 13-mm tubes
Scienceware polypropylene test tube rack for 26-30 mm tubes
Thermo Scientific Nalgene Unwire™ test tube rack for 13-mm tubes, w
Thermo Scientific Nalgene Unwire™ test tube rack for 13-mm tubes, o.
More Details
Atm Stainless Steel Testing Sieve
Buy Atm Stainless Steel Testing Sieve Items Including:
Receiving Pan for 12" Stainless Steel Bottom Sieves, Full Height
Receiving Pan with Fitted Rim for Nesting 12" Stainless Steel Sieves, (YO-59996-92)
Receiving Pan with Fitted Rim for Nesting 12" Stainless Steel Sieves, (YO-59996-93).
More Details
Corning Ph Meter 430 Replacement Electrode
Buy Corning Ph Meter 430 Replacement Electrode Items Including:
Replaceable electrode; glass body, refillable, no ATC
Free Chlorine Replacement Electrode
Free Chlorine Replacement Electrode for Self Cleaning
Thermo Scientific- Orion ROSS Replacement Electrode Fill Solution, (4)
Replacement electrode cartridge.
More Details
Ansi Flange Gasket
Buy Ansi Flange Gasket Items Including:
ANSI 150# Ff Gasket,epdm,1/2" (YO-30549-01).
More Details
500 Ml Kimax Kimble Filter Flask
Buy 500 Ml Kimax Kimble Filter Flask Items Including:
KIMAX filtering flask, 1000 mL
KIMAX filtering flask, 2000 mL, 1/ea
KIMAX filtering flask, 4000 mL, 1 ea
500 mL Kimax Amber Media Bottles
KIMAX baffle shake flask, 500 mL
More Details
Oil-Less Diaphragm Air Compressors
Buy Oil-Less Diaphragm Air Compressors Items Including:
Oilless Air Compressor, Diaphragm compressor pump, 0.52 cfm, 220/230 V (MOA-P101-CD)
Oilless Air Compressor, Diaphragm compressor pump, 1.1 cfm, 220/240 VA (DOA-P501-BN).
More Details
Separatory Funnel
Buy Separatory Funnel Items Including:
Cole-Parmer Separatory Funnel, 250mL, PP 2/pk (YO-06124-04)
Thermo Scientific Nalgene separatory funnel with polypropylene body an (YO-06126-10)
Scienceware Separatory Funnel Rack
Kimble Separatory Funnel, PTFE Stopcock, 125 mL 4/cs (YO-34505-44).
More Details
High Temperature Plastic Ball Valve
Buy High Temperature Plastic Ball Valve Items Including:
Ball Valve, 2-way, 1/8" NPT(F), PP with EPDM seals
Ball Valve, 2-way, 1/8" NPT(F), PP with Viton® seals
Ball Valve, 2-way, 1/8" NPT(F) x 1/8" NPT(M), PP with Viton® seals
Ball Valve, 2-way, 1/4" barb, PP with Viton® seals.
More Details
Viton Tubing
Buy Viton Tubing Items Including:
Viton Tubing, 1/16"ID x 1/8"OD, 25 ft/pack
Viton Tubing, 1/8"ID x 3/16"OD, 25 ft/pack
Viton Tubing, 3/16"ID x 1/4"OD, 25 ft/pack
Viton Tubing, 5/16"ID x 7/16"OD, 25 ft/pack.
More Details
Mixed Cellulose Ester Membrane
Buy Mixed Cellulose Ester Membrane Items Including:
Advantec Mixed Cellulose Ester Membranes, 0.30 Wp 47mm 100/pk
Advantec Mixed Cellulose Ester Membranes, 0.65 Wp 47mm 100/pk
Advantec Mixed Cellulose Ester Membranes, 0.80 Wp 37mm 100/pk
Advantec Mixed Cellulose Ester Membranes, 0.80 Wg 25mm 100/pk.
More Details
1 Micron Nylon Filter Bags
Buy 1 Micron Nylon Filter Bags Items Including:
General-purpose aluminum filter bag housing, 1-1/4" NPT(F) (156109)
General-purpose 304 stainless steel filter bag housing, 1-1/4" NPT(F) (156120)
General-purpose 304 stainless steel filter bag housing, 2" NPT(F) (156121).
More Details
70618
results
<<<Previous
16
17
18
19
20
21
22
23
24
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Thermo Scientific Thermolyne Atmosphere-Controlled Muffle Furnaces to 1 Micron Nylon Filter Bags
>
31125329-s-toc_2 31125329stoc2