Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 200
Thermo Scientific Barnstead Nanopure Water Purification
to
Dissolved Oxygen Meter Hach
Image
Product Detail
More Detail
Thermo Scientific Barnstead Nanopure Water Purification
Buy Thermo Scientific Barnstead Nanopure Water Purification Items Including:
Cartridge Pack for use with Thermo Scientific Barnstead NANOpure™ W (D50281)
Nanopure Life Science Systems (RO/Distilled Feed) Consumables Bundle (D502144)
Thermo Scientific Barnstead Replacement Filters for remote dispenser, (D3751).
More Details
Pyrex Media Bottle Cap
Buy Pyrex Media Bottle Cap Items Including:
Pyrex Brand 1395 media storage bottle w/screw cap, 25 mL
Pyrex Brand 1395 media storage bottle w/screw cap, 100 mL
Pyrex Brand 1395 media storage bottle w/screw cap, 250 mL
Pyrex Brand 1395 media storage bottle w/screw cap, 500 mL.
More Details
Smc Industries Solenoid Valves
Buy Smc Industries Solenoid Valves Items Including:
Solenoid Valve, 2-Way Normall Closed, 12V, with 1/8" Tube Connect (LVM10R1-6C-3)
Solenoid Valve 2-Way, Zero Differential Pressure, Normall Closed, 1/4 (VXZ22 30-02N-5DZ1-B)
Solenoid Valve, 2-Way Normall Closed, 24V, with 1/8" Tube Connect.
More Details
Bunsen Burner Gas Types
Buy Bunsen Burner Gas Types Items Including:
Bunsen burner, adjustable with threaded needle valve, natural gas (H-5885)
Bunsen burner, adjustable with threaded needle valve, LP gas (H-5890)
Bunsen burner, Tirrill burner, LP gas (H-6225)
Bunsen burner, high-temperature burner, LP gas (H-5600).
More Details
Admiral Vacuum Pumps
Buy Admiral Vacuum Pumps Items Including:
Cole-Parmer Vacuum Pump, 0.37 cfm, 20" Hg, 120 VAC
Cole-Parmer Vacuum Pump, 0.31 cfm, 21.3" Hg, 230 VAC
Air Admiral diaphragm vacuum/pressure pump, 0.37 cfm, 115 VAC
Air Admiral diaphragm vacuum/pressure pump, 0.31 cfm, 230 VAC.
More Details
2 Gallon Narrow Mouth Hdpe Bottle
Buy 2 Gallon Narrow Mouth Hdpe Bottle Items Including:
Thermo Scientific Nalgene Narrow-Mouth Economy Bottle, HDPE, 30 mL, 12 (YO-06035-07)
Thermo Scientific Nalgene Narrow-Mouth Economy Bottle, HDPE, 30 mL, 72 (YO-06035-09).
More Details
Cole-Parmer Brass Compression to Npt(M) Adapters
Buy Cole-Parmer Brass Compression to Npt(M) Adapters Items Including:
Compression male pipe adapter elbow, 1/8" NPT(M) x 1/8" OD, brass
Compression male pipe adapter elbow, 1/8" NPT(M) x 1/4" OD, brass
Compression pipe adapter, Brass, 1/2" x 3/8", Each (YO-31412-75).
More Details
Thin Wall Steel Tubing Fittings
Buy Thin Wall Steel Tubing Fittings Items Including:
316 Stainless Steel Tubing; dimensions, 1/16" ID, 1/8" OD, 1/32" wall; (YO-03300-05)
Compression Straight Union, Stainless Steel, 6mm OD Tubing (YO-31528-33)
Compression Straight Union, Stainless Steel, 10mm OD Tubing (YO-31528-35).
More Details
Cole-Parmer Un Spec (1h1) Tight-Head Drums
Buy Cole-Parmer Un Spec (1h1) Tight-Head Drums Items Including:
UN Specification (1H1) Tight-Head Drum, 30 Gal
(YO-06950-40).
More Details
Male to Female Luer Valve
Buy Male to Female Luer Valve Items Including:
1/8" NPT brass female/male mini manual ball valve (MV608-2)
1/4" NPT brass female/male mini manual ball valve (MV608-4)
Male luer x female luer coupler, PC, 25/pk (YO-45501-82)
Male luer x female luer coupler, Nylon, 25/pk (YO-45502-82).
More Details
2 Micron Filter Paper Cones
Buy 2 Micron Filter Paper Cones Items Including:
Advantec Grade No
2 Qualitative Filter Paper; 5.5 cm dia.; pack of 10
Advantec Grade No
2 Qualitative Filter Paper; 7.0 cm dia.; pack of 10
Advantec Grade No
More Details
Oakton Conductivity Standard Solution
Buy Oakton Conductivity Standard Solution Items Including:
Oakton conductivity solution, 12,880 µS
Oakton® conductivity solution, 1413 µS
Oakton® conductivity solution, 2764 µS
Oakton® conductivity solution, 2070 µS
Oakton® conductivity solution, 80,000 µS
More Details
Cole-Parmer Grip Tight Circuit Breaker Lockouts
Buy Cole-Parmer Grip Tight Circuit Breaker Lockouts Items Including:
Grip Tight, Single/Double Pole Circuit Breaker Lockout (KDD135).
More Details
Tube Extension Set Male Female Luer
Buy Tube Extension Set Male Female Luer Items Including:
316 SS fittings; female luer x male luer lock (YO-31507-21)
Nickel-plated brass fittings, female luer x male luer (YO-31507-50)
Side arm injection port, 2 female, 1 male slip luer (YO-31507-83)
Luer Tee, Female-Male-Female, PP, 25/pk (YO-45500-85).
More Details
Cole-Parmer Class a Sample Glass Vials
Buy Cole-Parmer Class a Sample Glass Vials Items Including:
Glass Vials PTFE/SILiCone SEPTA Clear 12 ml 200/pk (B7990-12)
Glass Vials PTFE/SILiCone SEPTA Clear 16 ml 200/pk (B7990-4)
Glass Vials PTFE/SILiCone SEPTA Clear 22 ml 200/pk (B7990-5)
Glass Vials PTFE/SILiCone SEPTA Amber 12 ml 200/pk (B7990-12A).
More Details
36 Inch Stainless Steel Temperature Probe
Buy 36 Inch Stainless Steel Temperature Probe Items Including:
Type J Heavy-duty stainless-steel Thermocouple probe with T-handle; 10 (050897A-J)
Type K Heavy-duty stainless-steel Thermocouple probe with T-handle; 10 (050897A-K)
Type T Heavy-duty stainless-steel Thermocouple probe with T-handle; 10 (050897A-T).
More Details
Nylon Straight Compression Unions
Buy Nylon Straight Compression Unions Items Including:
Compression union, Nylon, 1/4" 100 Pack (YO-30803-03)
Compression union, Nylon, 5/16" 100 Pack (YO-30803-04)
Compression union, Nylon, 3/8" 100 Pack (YO-30803-05)
Compression union, Nylon, 1/2" 100 Pack (YO-30803-06).
More Details
Proguard Hand Cream
Buy Proguard Hand Cream Items Including:
Proguard Professional Hand Cream, 3 oz, cs/12
Proguard Professional Hand Cream, 3 oz
Proguard Professional Hand Cream, 16 oz, cs/6.
More Details
10mm Tube Compression Fittings
Buy 10mm Tube Compression Fittings Items Including:
PFA compression threaded adapter, 10mm tubing OD x 3/8" male NPT (YO-31321-45)
Compression To Thread Adapter, Stainless Steel, 10mm OD Tubing X 1/4" (YO-31528-07)
Compression Straight Union, Stainless Steel, 10mm OD Tubing (YO-31528-35).
More Details
Probe Adapters, Connectors, and Extension Cables
Buy Probe Adapters, Connectors, and Extension Cables Items Including:
RTD Probe Adapter; pins are 0.31" apart (93400-50)
RTD Probe Adapter; pins are 0.44" apart (93400-52)
RTD Probe Extension Cable; 25'L (041398B-25)
Adapter, ANSI 3-pin male miniconnector to Cole-Parmer 3-pin male (011900M).
More Details
Portable Air Compressor Tanks
Buy Portable Air Compressor Tanks Items Including:
Professional series air compressor, 7.1 cfm, 20 gallon portable tank,.
More Details
Supplied Air Systems
Buy Supplied Air Systems Items Including:
Allegro One Man Full Face Piece Supplied Air System with 50 foot Hose.
More Details
Rnase Free Screw Cap Centrifuge Tubes
Buy Rnase Free Screw Cap Centrifuge Tubes Items Including:
15 mL conical bottom disposable plastic centrifuge tubes, screw cap/ri (3219P)
15 mL conical bottom disposable plastic centrifuge tubes, screw cap/ri (3215P)
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 5 ml, 125 (99502-5).
More Details
Ptfe Capsule Filter
Buy Ptfe Capsule Filter Items Including:
Disposable High-Area Capsule Filters, PTFE Membrane Capsules; pore siz (YO-29819-00)
Whatman™ Disposable Capsule Filters, PTFE Membrane; pore size, 0.2 (YO-29700-00)
Whatman™ Disposable Capsule Filters, PTFE Membrane; pore size, 1.0 (YO-29700-02).
More Details
Ph Indicator Papers
Buy Ph Indicator Papers Items Including:
Universal pH Test Paper; pH Range 1 to 11; 9 mm x 6 m Roll
Litmus Red Test Paper, 10 Booklets of 20 Strips
pHydrion 1-12 Paper Strips
Hydrion quaternary ammonium test paper, 15-ft roll, box of 10
Hydrion chlorine test paper, 15-ft roll, box of 10.
More Details
Hydroxide Chemicals, Reagents, & Lubricants
Buy Hydroxide Chemicals, Reagents, & Lubricants Items Including:
SODIUM HYDROXIDE NF/EP/BP/FCC (YO-88081-49)
LITHIUM HYDROXIDE MONOHYDR 25G (YO-88213-23)
ALUMINUM HYDROXIDE POWDER 25G (YO-88234-23)
POTASSIUM HYDROXIDE REAG 2.5KG (YO-88362-44)
POTASSIUM HYDROXIDE REAG 500G (YO-88362-46).
More Details
Fluoroware 2 Way Valve 1 4
Buy Fluoroware 2 Way Valve 1 4 Items Including:
PTFE 2-Way Stopcock, 1/4" Orifice, 1/4" NPT(F) connections (YO-98515-82)
PP Three-Way, Two-Position Elliptic Valve, 1/4" NPT(F) connectors (223-PP-1)
PVDF Three-Way, Two-Position Elliptic Valve, 1/4" NPT(F) connectors (223-PV-1).
More Details
Blue or Purple Latex Free Gloves
Buy Blue or Purple Latex Free Gloves Items Including:
Microflex ColorTouch Pink Powder-Free Gloves, Extra small
AURELIA™ Velocity™ Powder-Free Latex Gloves; size, small
AURELIA™ Velocity™ Powder-Free Latex Gloves; size, medium
AURELIA™ Velocity™ Powder-Free Latex Gloves; size, large.
More Details
Gravity Cup
Buy Gravity Cup Items Including:
Specific Gravity Cup, 83.2 mL capacity, 0.5% volume tolerance, for use (PV-9654)
Specific gravity cup, 8.32 mL capacity, 1.2% volume tolerance, for use (WG-SS-8.32)
Specific gravity cup, 83.2 mL capacity, 0.5% volume tolerance, for use (WG-SS-83.2).
More Details
Dissolved Oxygen Meter Hach
Buy Dissolved Oxygen Meter Hach Items Including:
Hach SensION + Dissolved Oxygen Meter Kit
YSI splashproof dissolved oxygen meter with probe, 25-ft cable, model
YSI Model 550A Dissolved Oxygen Meter, 12-ft (3.7-m) probe cables
YSI DO 200 Dissolved Oxygen Meter Only
Extech Dissolved Oxygen Meter.
More Details
70618
results
<<<Previous
196
197
198
199
200
201
202
203
204
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
100
110
120
130
140
150
160
170
180
185
186
187
188
189
190
191
192
193
194
195
196
197
198
199
200
201
202
203
204
205
206
207
208
209
210
211
212
213
214
220
230
240
250
260
270
280
290
300
400
500
600
700
800
900
1,000
1,100
2,000
2,354
Thermo Scientific Barnstead Nanopure Water Purification to Dissolved Oxygen Meter Hach
>
31125329-s-toc_2 31125329stoc2