Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 2000

Plastic Sealing   to   Work Hoods

Image
  Product Detail
More Detail

Plastic Sealing

Buy Plastic Sealing Items Including:
  • All-Plastic Quick Disconnect Sealing Cap, PC, 5/pack (MPC32003)
  • Snap-Seal™ Plastic Sample Containers, Translucent PP, 7/16 oz
  • Snap-Seal™ Plastic Sample Containers, Translucent PP, 1-1/2 oz
  • Snap-Seal™ Plastic Sample Containers, Translucent PP, 4 oz.

Dew Point Temp

Buy Dew Point Temp Items Including:
  • Digital Psychrometer, RH, Temp, Dew Point, & Wet Bulb (SAM990DW).

Ss Boxes

Buy Ss Boxes Items Including:
  • Stainless Steel (SS) Hose Clamps, 11/16 x 1-1/4", box of 10
  • Stainless Steel (SS) Hose Clamps, 13/16 x 1-3/4", box of 10
  • Stainless Steel (SS) Hose Clamps, 1-5/16 x 2-1/4", box of 10.

4 Probes Resistivity Measurement

Buy 4 Probes Resistivity Measurement Items Including:
  • Oakton® Acorn™ TDS 6 Meter with probe (YO-35606-15)
  • Oakton® Acorn™ SALT 6 Meter with probe (YO-35606-20)
  • Vane Probe Thermoanemometer with 2-3/4" (70mm) vane.

Utility 300

Buy Utility 300 Items Including:
  • 0 to 300 psi Utility Gauge, 1 1/2" Dial, Back Connection
  • 0 to 300 psi Utility Gauge, 2" Dial, Bottom Connection
  • 0 to 300 psi Utility Gauge, 2 1/2" Dial, Bottom Connection
  • Low-Cost Utility Pressure Gauge, 0-300 psi (KC102158300).

Large Centrifuges

Buy Large Centrifuges Items Including:
  • Large Capacity Variable-Speed Centrifuge; 110V (YO-17451-00)
  • Large Capacity Variable-Speed Centrifuge; 220V (YO-17451-05)
  • High-Speed Programmable Refrigerated Universal Centrifuge; 120V (YO-17303-10)
  • Swing-out Rotors; 16 place x 15mL (YO-17451-60).

Peristaltic Tubing Set

Buy Peristaltic Tubing Set Items Including:
  • Tube Set, PharMed, 0.51 mm ID; Set of 4
  • Tube Set, PharMed, 1.03 mm ID; Set of 4
  • Tube Set, PharMed, 1.52 mm ID; Set of 4
  • Tube Set, PharMed, 2.06 mm ID; Set of 4
  • Pumps, Metering Pumps Replacement Tube Set for Mid-Flow Range Cole-Par (YO-74204-01).

Frost Covers

Buy Frost Covers Items Including:
  • Semimicro cuvettes with frosted thick wall; cover top; glass; 1.4 mL c
  • Semimicro cuvettes with frosted thick wall; cover top; quartz; 1.4 mL
  • Cole-Parmer Brand Slides; frosted; one gross (144 slides)
  • Cole-Parmer Brand Slides; frosted; 10 gross/case.

Compression Hoses

Buy Compression Hoses Items Including:
  • Compression reducing union, Pharmafluor™ PTFE, 1/4" OD x 1/8" OD (YO-06379-11)
  • Compress to male threaded adapter, Pharmafluor PTFE, 1/16" OD x 1/8" N
  • Pharmafluor Compression To Male NPT Thread Elbow, PFA, 3/8 X 1/4 (YO-31001-52).

Cole Bacteria Test Kits

Buy Cole Bacteria Test Kits Items Including:
  • Watersafe Bacteria Test Kit
  • Listeria Swabcheck kit, 25 tests (YO-14019-00)
  • E
  • coli Swabcheck kit, 125 tests (YO-14019-10)
  • Botulinum toxin detection kit (YO-36108-14)

Gm Quick Connects Thread Size

Buy Gm Quick Connects Thread Size Items Including:
  • Push-to-connect threaded elbow adapters, polypropylene, 1/4" x 1/8" NP (YO-34007-62)
  • Push-to-connect threaded elbow adapters, polypropylene, 1/4" x 1/4" NP (YO-34007-63).

Hepa Systems

Buy Hepa Systems Items Including:
  • Primary HEPA Filter Package with Carbon Module for Air Cleaning System (YO-33703-04)
  • Replacement Prefilters for HEPA Filter for Air Cleaning Systems, Pack (YO-33703-12)
  • HEPA Filter for use with Ductless Air Filter System (YO-33703-43).

Portable Moisture Meter

Buy Portable Moisture Meter Items Including:
  • LabSwift Portable Water Activity Instrument
  • HART 1552A-12 Handheld Thermometer, Fixed RTD, –80 To 300C, 1/4x12
  • Hart Carrying Case, For Hart 1551 / 1552
  • HART 1551A-9 Handheld Thermometer, Fixed RTD, –50 To 160C, 3/16x9.

Labware Plastics

Buy Labware Plastics Items Including:
  • ESD Plastic Tweezers, Flat Round Tip, Pattern 702A
  • ESD Plastic Tweezers, Ultra Fine Tip, Pattern 705
  • ESD Plastic Tweezers, Flat Fine Tip, Pattern 709
  • Curved: Smooth Rounded Tips 2AB (18.514)
  • Polypropylene tri-corner beaker assortment pack (80091).

General Purpose Cleaner

Buy General Purpose Cleaner Items Including:
  • Micro A07 Citric Acid Cleaner case of 12 1L bottles (YO-18104-01)
  • Liqui-Nox cleaner for general purpose cleaning, 1 quart bottle, case o
  • Liqui-Nox cleaner for general purpose cleaning, 1 quart bottle
  • Sterile 70% Isopropanol 16 oz Polybottles 12/CS.

Thermistor Food

Buy Thermistor Food Items Including:
  • Thermistor Food Probe (0613 2411)
  • 400 Series Thermistor small flexible probe for frozen food packages an (423)
  • Small flexible probe, for forzen food packages and cuvettes
  • Nylon and (08491-17).

Lab Glassware Bath

Buy Lab Glassware Bath Items Including:
  • Rotary Evaporator with Diagonal glassware, water bath, 115V/60Hz (YO-28622-05)
  • Rotary Evaporator with vertical glassware, water bath, 115V/60Hz (YO-28622-25)
  • Rotary Evaporator with cold-trap glassware, water bath, 115V/60Hz (YO-28622-45).

Cole-Parmer Dataloggers

Buy Cole-Parmer Dataloggers Items Including:
  • Dual-channel multifunction datalogger
  • NIST-Traceable Calibration; Datalogger (YO-17002-20)
  • Deluxe modular handheld datalogger (0563 4001)
  • Replacement RS-232 cable for datalogger kit
  • Cole-Parmer Heavy-Duty Hot-Wire Thermoanemometer with Datalogger.

Excel Microbial Analysis

Buy Excel Microbial Analysis Items Including:
  • Lazy-L-Spreader™, disposable, 500/case;10/pak (YO-14201-00).

Excel Scientific Lazy

Buy Excel Scientific Lazy Items Including:
  • Lazy-L-Spreader™, disposable, 500/case;10/pak (YO-14201-00).

Measuring Motor Torque

Buy Measuring Motor Torque Items Including:
  • Esm:Motorized Test Stand 110V.

Lab Systems

Buy Lab Systems Items Including:
  • Lab-Tek™ Chambered Cover Glass System; material, glass; number of w (YO-01838-10)
  • Lab-Tek™ Chambered Cover Glass System; material, glass; number of w (YO-01838-11)
  • Lab-Tek™ Chambered Cover Glass System; material, glass; number of w (YO-01838-12).

Test Product

Buy Test Product Items Including:
  • Test Products International Handheld Particle Counter
  • Ammonia Nitrogen Reagent Set, Salicylate Method, 0-0.50 ppm, 100 Tests
  • Refill kit, Phenol test, for 00290-30
  • Refill kit, Copper, total soluble test, for 00290-50
  • PFA Test Tubes With Caps, 5 mL.

High Pressure Equipment

Buy High Pressure Equipment Items Including:
  • Advantec High-Pressure Stainless Steel Filter Holder, 47 mm
  • Autofill Valve Box (High Pressure) for Cole-Parmer Syringe Pump, Infus (788320) (YO-74903-71)
  • Electrode, pH, High Pressure
  • Additel 919 High Pressure Pneumatic Test Pump Carrying Case.

0.5 Lpm Pump

Buy 0.5 Lpm Pump Items Including:
  • Cole-Parmer Polystat recirculator with force/suction pump, 21 lpm, 250 (YO-13042-05).

10 Bath

Buy 10 Bath Items Including:
  • Open Refrigerated Bath, AC-150 controller, 17-30 liter capacity, -10 t (YO-12145-10)
  • Polystat economical constant temperature bath, 120 VAC
  • Cannon standard constant temperature bath, 120 VAC
  • Aquabath™ 10L Analog Utility Bath - 120V (YO-03710-13).

Outdoor Temperature

Buy Outdoor Temperature Items Including:
  • Indoor/Outdoor Thermometer with settable Hi/Low audible alarm
  • Indoor/Outdoor Thermometer (4126CP)
  • Extech 445713 Big Digit Hygro-Thermometer Indoor/Outdoor (445713)
  • 12" Dial Metal Thermometer -60F to 120F and -50C to 50C
  • Suitable for (49562J).

Wall Mounted Thermometers

Buy Wall Mounted Thermometers Items Including:
  • Thermometers with Time/Date and Wire Probe (4105CP)
  • Dual Channel Thermometer with 2 Wire Probes (4307CP)
  • Jumbo Refrigerator/freezer thermometer with a 5ml bottle that fits in (4627CP)
  • Jumbo Refrigerator/freezer thermometer with Wire Probe (4148CP).

Water Resistant Materials

Buy Water Resistant Materials Items Including:
  • +GF+ Signet Conductivity/Resistivity Cell, 316SS, K = 0.01, reversible (3-2819-1)
  • +GF+ Signet Conductivity/Resistivity Cell, 316SS, K = 0.1, reversible (3-2820-1)
  • +GF+ Signet Conductivity/Resistivity Cell, 316SS, K = 10.0, 3/4" NPTM (3-2822-1).

Work Hoods

Buy Work Hoods Items Including:
  • Stainless Steel Work Surface, 5-ft For 35.5"w Xpert Systems (YO-09109-86)
  • Stainless Steel Work Surface, 6-ft For 35.5"w Xpert Systems (YO-09109-88)
  • Laconco Work surface, spillstopper, 48"
  • Laconco Work surface, spillstopper, 60".



Plastic Sealing to Work Hoods

>