Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 21
Sol-Vex Reusable Nitrile Gloves
to
Pyrex Round Bottom Flask
Image
Product Detail
More Detail
Sol-Vex Reusable Nitrile Gloves
Buy Sol-Vex Reusable Nitrile Gloves Items Including:
Ansell Edmont Sol-Vex Nitrile Gloves, Unlined, size 9, 22-mil, 18"L, 1
Ansell Edmont Sol-Vex Nitrile Gloves; Unlined; size 10; 11-mil; 13"L; (37-185 117301)
Ansell Edmont Sol-Vex Nitrile Gloves, Unlined, size 9, 11-mil, 13"L, 1.
More Details
Oil Less Rotary Vane Vacuum Pumps
Buy Oil Less Rotary Vane Vacuum Pumps Items Including:
Vacuubrandreg Rotary Vane Vacuum Pump, 1.6 cfm, 1.5 X 10-3/sup (698123)
Vacuubrandreg Rotary Vane Vacuum Pump, 1.6 cfm, 1.5 X 10-3/sup (698120)
Vacuubrandreg Rotary Vane Vacuum Pump, 4.0 cfm, 1.5 X 10-3/sup (698133).
More Details
Digital Pressure Gauges
Buy Digital Pressure Gauges Items Including:
Ashcroft Digital Pressure Gauge, 0 to 1000 psi, 1/4" NPT(M) Process co (D1005PS2)
Digital Pressure Gauge/controller, 1 PSIG; 1/8" NPT(F) Process Connect
Digital Pressure Gauge/controller, 5 PSIG; 1/8" NPT(F) Process Connect.
More Details
Kynar Fittings
Buy Kynar Fittings Items Including:
Adapter, Kynar, 1/2" mini to 1/2" to female luer lock
Barbed fitting kits, BSPT, adapters/elbows/tees, Kynar
Threaded barb small adapter kits; Kynar material (set of 1)
adapter; female luer lock to two-way valve; Kynar body and PP insert.
More Details
Mixer Mill
Buy Mixer Mill Items Including:
10-mL Mill Jar; stainless steel (YO-04182-31)
25-mL Mill Jar; stainless steel (YO-04182-32)
35-mL Mill Jar; stainless steel (YO-04182-33)
50-mL Mill Jar; stainless steel (YO-04182-34)
10-mL Mill Jar; Agate (YO-04182-38)
More Details
500 Ml Erlenmeyer Flask
Buy 500 Ml Erlenmeyer Flask Items Including:
Universal 500-mL Erlenmeyer flask clamp
Dedicated Plateform complete with 500 ml Erlenmeyer flask clamps
500 ml Erlenmeyer Flask Clip
PYREX Brand 4995 flask; 500 mL, case of 6
PYREX Brand 4980 flask; 500 mL, case of 36
More Details
Pinnacle Ph Electrodes
Buy Pinnacle Ph Electrodes Items Including:
Replacement three-in-one pH electrode, gel-filled junction; 6-ft cable
Replacement high-performance pH electrode, combination, glass body, BN
Three-in-one pH/reference/temperature electrode, epoxy body, BNC for m (476436P)
ATC Probe (477968).
More Details
To 1 Bspp Female Reducer
Buy To 1 Bspp Female Reducer Items Including:
316 SS Female Reducer, 1/4" x 1/8" NPT(F) (YO-31211-10)
316 SS Female Reducer, 1/2" x 1/8" NPT(F) (YO-31410-13)
Threaded adapter, stainless steel, 1/8" female NPT to 1/8" male BSPP (YO-31830-50).
More Details
A&D Weighing Food and Portion Scales
Buy A&D Weighing Food and Portion Scales Items Including:
A&D SJ Series Digital Portion Scale, 2000 G X 1 G
A&D SJ Series Digital Portion Scale, 12 Kg X 0.005 Kg
A&D Portable Stainless Steel Washdown Scale, 1000 g (SK-1000WP)
A&D Portable Stainless Steel Washdown Scale, 2000 g (SK-2000WP).
More Details
Cryobox Cryogenic Storage Boxes and Racks
Buy Cryobox Cryogenic Storage Boxes and Racks Items Including:
Thermo Scientific Nalgene Polycarbonate CryoBox storage boxes for 25 (
Thermo Scientific Nalgene Polycarbonate CryoBox storage boxes for 100
Cardboard Cryobox 5 1/4 X 5 1/4 X 2 (R3015)
Cardboard Cryobox 5 1/4 X 5 1/4 X 2 w/100 Place Insert (R3027).
More Details
Bspt Barb Fittings
Buy Bspt Barb Fittings Items Including:
Barbed fitting kits, BSPT, adapters/elbows/tees, Natural PP (YO-31514-30)
Barbed fitting kits, BSPT, adapters/elbows/tees, Black Nylon (YO-31514-29)
Barbed fitting kits, BSPT, adapters/elbows/tees, HDPE (YO-31514-32).
More Details
5 Firematic Check Valve
Buy 5 Firematic Check Valve Items Including:
Bel-Art LDPE Check Valve, 5/16" to 1/4" hose barb
PP Miniature Check Valve, 5/16" hose barb, Viton diaphragm.
More Details
Miniature Vacuum Pumps
Buy Miniature Vacuum Pumps Items Including:
PUMP, VACUUM PUMP WITH BRUSHLESS DC MOTOR, 1.5 LPM, 12 VDC, GAS ONLY,
PUMP, VACUUM PUMP WITH BRUSHLESS DC MOTOR, 1.5 LPM, 24 VDC, GAS ONLY,
Miniature venturi vacuum pump, 0.5 cfm, 28.0
Compact/OEM bellows pressure pump, 0.5 LPM 5.8 psi, 3 VDC.
More Details
Kimax Glass Beakers
Buy Kimax Glass Beakers Items Including:
Kimax blue-coded Griffin beakers; 50 mL, 12/cs
Kimax blue-coded Griffin beakers; 100 mL, 12/cs
Kimax heavy-duty beakers; 250-mL, 48/cs
Kimax heavy-duty beakers; 400-mL, 48/cs
Kimax heavy-duty beakers; 1000-mL, 24/cs.
More Details
Sieve Cleaning Brushes
Buy Sieve Cleaning Brushes Items Including:
Rectangular Sieve Brush for 100 Mesh Sieves and Coarser, Brass Bristle.
More Details
Acid Storage Cabinets
Buy Acid Storage Cabinets Items Including:
SciMatCo Full-Size Acid Storage Cabinet, twenty four 2.5 L bottle capa
SciMatCo Full-Size Acid Storage Cabinet with Full Lining, thirty 2.5 L
SciMatCo HDPE Benchtop Acid Storage Cabinet, six 2.5 L bottle capacity.
More Details
Solenoid-Operated 2 and 3-Way Pinch Valves
Buy Solenoid-Operated 2 and 3-Way Pinch Valves Items Including:
3-way pinch valve, 12 VDC, 1/16" x 1/8" (SCH384A002- 12VDC)
3-way pinch valve, 24 VDC, 1/16" x 1/8" (SCH384A002- 24VDC)
Two-way normally closed solenoid pinch valve; 12 VDC, 1/16" ID x 3/16" (100P2NC-12-03).
More Details
Ika Big Squid Stirrer
Buy Ika Big Squid Stirrer Items Including:
IKA Digital Big Squid Stirrer 100-240V
Ika Digital Color Squid (white) 110-240V.
More Details
Stainless Overmolded Luer Fittings
Buy Stainless Overmolded Luer Fittings Items Including:
Luer fittings, 316 SS fittings, male luer lock x 1/16" hose barb (YO-31507-26)
Luer fittings, 316 SS fittings, female luer lock x 1/16" hose barb (YO-31507-29)
Stainless steel stopcocksFemale luer x female luer x male luer loc (V5880003).
More Details
Steel Luer Fittings
Buy Steel Luer Fittings Items Including:
Luer fittings, 316 SS fittings, female luer lock x 1/16" hose barb (YO-31507-29)
316 SS fittings; male slip plug (YO-31507-22)
316 SS fittings; male luer lock plug (YO-31507-23)
316 SS fittings; female luer x 1/8" hose barb (YO-31507-28).
More Details
Pharmed Tubing
Buy Pharmed Tubing Items Including:
Tubing sets, PharMed BPT with 2 welded stops, 400 mm length, 0.25 mm I (SC0740)
2-Stop PharMed BPT Tubing, 0.25mm, 12/pack (95713-12)
2-Stop PharMed BPT Tubing, 0.51mm, 12/pack (95713-18)
2-Stop PharMed BPT Tubing, 0.89mm, 12/pack (95713-26).
More Details
Nilfisk Vacuum Bags
Buy Nilfisk Vacuum Bags Items Including:
Replacement vacuum bags, pack of 5
Replacement Dust Bags for Nilfisk Backpack HEPA Vacuum, 5/pack.
More Details
Nilfisk Vacuum Cleaner Bags
Buy Nilfisk Vacuum Cleaner Bags Items Including:
Nilfisk HEPA Back-Mounted Vacuum, Replacement Bags (22198000)
Replacement Dust Bags for Nilfisk Backpack HEPA Vacuum, 5/pack.
More Details
Cleanroom Vinyl Gloves
Buy Cleanroom Vinyl Gloves Items Including:
QRP Vinyl Cleanroom Gloves, small, 100/bag
QRP Vinyl Cleanroom Gloves, medium, 100/bag
QRP Vinyl Cleanroom Gloves, large, 100/bag.
More Details
Cole-Parmer Quiet Vacuum/Pressure Pumps
Buy Cole-Parmer Quiet Vacuum/Pressure Pumps Items Including:
Vacuum/Pressure Pump, 2.0 CFM 115V
Quiet Pressure Pump, 2.3 CFM, 3.0psi max, 115VAC.
More Details
Ptfe Npt(M) Threaded Plugs
Buy Ptfe Npt(M) Threaded Plugs Items Including:
Threaded Plug, PTFE, 1/8" NPT(M) (F-PL01)
Threaded Plug, PTFE, 1/4" NPT(M) (F-PL02)
Threaded Plug, PTFE, 3/8" NPT(M) (F-PL03)
Threaded Plug, PTFE, 1/2" NPT(M) (F-PL04)
Threaded Plug, PTFE, 3/4" NPT(M) (F-PL06).
More Details
Accuflame Tirrill Burners
Buy Accuflame Tirrill Burners Items Including:
Bunsen burner, Tirrill burner, natural gas (H-6220).
More Details
Honeywell Chart Recorder Paper
Buy Honeywell Chart Recorder Paper Items Including:
Replacement Chart Paper for Honeywell DR4300 10" Process Recorders, 10.
More Details
Multimeter Calibration Services
Buy Multimeter Calibration Services Items Including:
NIST-Traceable Calibration; Multimeter, Handheld 4.5-digit (YO-17090-00)
NIST-Traceable Calibration; Benchtop Multimeter, 4.5-digit (YO-17090-02)
NIST-Traceable Calibration; Handheld Multimeter, 5.5-digit (YO-17090-40).
More Details
Pyrex Round Bottom Flask
Buy Pyrex Round Bottom Flask Items Including:
PYREX Brand 4320 round-bottom flask; 1000 mL, pack of 1
PYREX Brand 4320 round-bottom flask; 2000 mL, pack of 1
PYREX Brand 4320 round-bottom flask; 5000 mL, pack of 1
PYREX Brand 4320 round-bottom flask; 1000 mL, case of 12.
More Details
70618
results
<<<Previous
17
18
19
20
21
22
23
24
25
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
40
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Sol-Vex Reusable Nitrile Gloves to Pyrex Round Bottom Flask
>
31125329-s-toc_2 31125329stoc2