Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 23
Stopcocks with Luer Connections
to
Bleach Water Testing Strips
Image
Product Detail
More Detail
Stopcocks with Luer Connections
Buy Stopcocks with Luer Connections Items Including:
Stopcocks with Luer Connections; 3-way; male lock; 5 flow pattern; 100 (YO-30600-07)
Stopcocks with Luer connection, male slip, 4-way, 6 flow pattern; 100/ (YO-30600-08)
Y-stopcock
2 female and 1 male connections, nickel-plated brass (YO-31507-15).
More Details
Thermo Scientific Nalgene Pipettes and Pipettors
Buy Thermo Scientific Nalgene Pipettes and Pipettors Items Including:
Thermo Scientific Nalgene ® Pipette Cleaning Baskets for 06172-00 (DS5241-0040)
Thermo Scientific Nalgene ® Pipette Cleaning Baskets for 06176-00 (DS5241-0050)
Thermo Scientific Nalgene ® Pipette Cleaning Jars, 20.25" H (5242-0040).
More Details
Hard Water Test Strips Delonghi
Buy Hard Water Test Strips Delonghi Items Including:
LaMotte Hardness Test Strips, 0 to 180 ppm
LaMotte Free Chlorine Test Strips, 0 to 10 ppm
LaMotte Free and Total Chlorine Test Strips, 0 to 10 ppm
LaMotte Hydrogen Peroxide Test Strips, 0 to50 ppm
EM Quant Total Hardness Test Strips, 0 to 370 ppm.
More Details
Tyvek Lab Coats
Buy Tyvek Lab Coats Items Including:
Tyvek Lab Coat, Medium
Tyvek Lab Coat, Large.
More Details
Thermo Scientific Nalgene Lowboy Low-Profile Carboys
Buy Thermo Scientific Nalgene Lowboy Low-Profile Carboys Items Including:
Thermo Scientific Nalgene Lowboys™ high-density polyethylene low-pr (2323-0008)
Thermo Scientific Nalgene Lowboys™ high-density polyethylene low-pr (2323-0015)
Thermo Scientific Nalgene Lowboys™ polypropylene low-profile carboy (2324-0008).
More Details
Thermo Orion Ph 550a
Buy Thermo Orion Ph 550a Items Including:
Thermo Scientific Orion pH electrode, glass body, refillable, single-j (9102BNWP)
Thermo Scientific Orion pH electrode, glass body, refillable, single-j (9103BNWP)
Thermo Scientific Orion pH electrode, glass body, refillable, single-j (9104BNWP).
More Details
Cole-Parmer Plastic Clamp Ties
Buy Cole-Parmer Plastic Clamp Ties Items Including:
Plastic Clamp Ties, White, 100/pack (YO-95615-00).
More Details
Balance Draft Shields
Buy Balance Draft Shields Items Including:
Cole-Parmer Balance Draft Shield, Small 13"W x 13-1/2"H x 14-3/4"D (small)
Cole-Parmer Balance Draft Sheild, Medium 15”W x 15-1/2”H x 16-3/4”D.
More Details
Glass Volumetric Flasks
Buy Glass Volumetric Flasks Items Including:
PYREX Brand 5640 Volumetric Flask; 100 mL, pack of 6
PYREX Brand 5640 Volumetric Flask; 200 mL, pack of 6
PYREX Brand 5640 Volumetric Flask; 1000 mL, pack of 1
PYREX Brand 5640 Volumetric Flask; 2000 mL, pack of 1.
More Details
Thermo Scientific Nalgene Carboys with Spigots
Buy Thermo Scientific Nalgene Carboys with Spigots Items Including:
Thermo Scientific Nalgene quick-action Tefzel ETFE spigot
Thermo Scientific Nalgene Replacement polypropylene cap, 24 mm
Thermo Scientific Nalgene Replacement polypropylene cap, 28 mm
Thermo Scientific Nalgene Replacement polypropylene cap, 100 mm.
More Details
Polypropylene Beakers
Buy Polypropylene Beakers Items Including:
Scienceware Tapered Polypropylene Beaker, 100 mL
Scienceware Tapered Polypropylene Beaker, 150 mL
Scienceware Tapered Polypropylene Beaker, 250 mL
Scienceware Tapered Polypropylene Beaker, 400 mL
Scienceware Tapered Polypropylene Beaker, 1000 mL.
More Details
Digital Pressure and Vacuum Gauges
Buy Digital Pressure and Vacuum Gauges Items Including:
Digital Pressure Gauge/controller With Dual-valves, 1 PSIG; 1/8" NPT(F
Digital Pressure Gauge/controller With Dual-valves, 5 PSIG; 1/8" NPT(F
Digital Pressure Gauge/controller With Dual-valves, 15 PSIG; 1/8" NPT(.
More Details
Compression Fitting 2 Steel Rubber
Buy Compression Fitting 2 Steel Rubber Items Including:
Compression Union, Stainless Steel, 3/4" x 1/2", Each (YO-31406-55)
Compression Fitting, Reducing Union, 316 SS, 1/2" x 1/4" OD (YO-31406-27)
Compression Fitting, Reducing Union, 316 SS, 1/2" x 3/8" OD (YO-31406-28).
More Details
Ammonium Hydroxide
Buy Ammonium Hydroxide Items Including:
AMMONIUM HYDROXIDE ACS 2.5L (YO-88053-91)
AMMONIUM HYDROXIDE 25% 2.5LT (YO-88252-97)
AMMONIUM HYDROXIDE REAG 2.5L (YO-88360-61).
More Details
Glass Bead Sterilizers
Buy Glass Bead Sterilizers Items Including:
Dry Glass Bead Sterilizer, 3" D x 1-3/4" dia sterilizing well; 110 VAC (YO-10779-00)
Dry Glass Bead Sterilizer, 3" D x 1-3/4" dia sterilizing well; 220 VAC (YO-10779-05).
More Details
Inch Pipe Check Valve
Buy Inch Pipe Check Valve Items Including:
PVC Check Valve, 1/4" NPT(F) connections (CKD025V-PV)
PVC Check Valve, 1/2" NPT(F) connections (CKM050V-PV)
PP Check Valve, 1/4" NPT(F) connections (CKD025V-PP)
PTFE Check Valve, 1/4" NPT(F) connections (CKD025V-TF).
More Details
Jacketed Pipe Check Valve
Buy Jacketed Pipe Check Valve Items Including:
PVC Check Valve, 1/4" NPT(F) connections (CKD025V-PV)
PVC Check Valve, 1/2" NPT(F) connections (CKM050V-PV)
PP Check Valve, 1/4" NPT(F) connections (CKD025V-PP)
PTFE Check Valve, 1/4" NPT(F) connections (CKD025V-TF).
More Details
Piping Check Valves
Buy Piping Check Valves Items Including:
PVC Check Valve, 1/4" NPT(F) connections (CKD025V-PV)
PVC Check Valve, 1/2" NPT(F) connections (CKM050V-PV)
PP Check Valve, 1/4" NPT(F) connections (CKD025V-PP)
PTFE Check Valve, 1/4" NPT(F) connections (CKD025V-TF).
More Details
Plastic 3 Way Ball Valve
Buy Plastic 3 Way Ball Valve Items Including:
Low-Cost PVC Two-Way Ball Valve, 3/4" NPT(F) connectors (TJH-002)
Ball Valve, 2-way, 3/8" barb, PVC with EPDM seals (YO-01377-42)
Ball Valve, 2-way, 3/8" barb, PVC with Viton® seals
Ball Valve, 2-way, 3/8" barb, with EPDM seals.
More Details
Pinnacle Ph Electrodes (Formerly Corning
Buy Pinnacle Ph Electrodes (Formerly Corning Items Including:
Replacement three-in-one pH electrode, gel-filled junction; 6-ft cable
Replacement high-performance pH electrode, combination, glass body, BN
Three-in-one pH/reference/temperature electrode, epoxy body, BNC for m (476436P)
ATC Probe (477968).
More Details
Stainless Steel Rubber Hose Barb Fittings
Buy Stainless Steel Rubber Hose Barb Fittings Items Including:
Female Cam X 3/4-in Hose Barb Adapter, SS (QC-C075-SS)
Barbed Y-Connector, Stainless Steel, 3/16
Barbed Y-Connector, Stainless Steel, 5/16
Barbed Y-Connector, Stainless Steel, 3/8
Straight Connector, Stainless Steel, 1/8", Each (YO-30620-01).
More Details
200 Ml Plastic Volumetric Flask
Buy 200 Ml Plastic Volumetric Flask Items Including:
Volumetric Flask, PMP, 25mL, 1/pk (YO-06110-32)
Volumetric Flask, PMP, 50mL, 1/pk (YO-06110-34)
Volumetric Flask, PMP, 100mL, 1/pk (YO-06110-36)
Volumetric Flask, PMP, 250mL, 1/pk (YO-06110-38)
Volumetric Flask, PMP, 500mL, 1/pk (YO-06110-40).
More Details
Chemistry Supplies Mortar and Pestle
Buy Chemistry Supplies Mortar and Pestle Items Including:
Cole-Parmer Mortar and Pestle Set, Porcelain; 80ML, Each
Cole-Parmer Mortar and Pestle Set, Porcelain; 500ML Each
Mortar and Pestle; Agate, 125ML, Each
Agate Mortar and Pestle, 25 mL (YO-04010-20)
Glass Mortar & Pestle Set, 50ml (YO-34500-51).
More Details
Lab Supplies Mortar and Pestle
Buy Lab Supplies Mortar and Pestle Items Including:
Cole-Parmer Mortar and Pestle Set, Porcelain; 80ML, Each
Cole-Parmer Mortar and Pestle Set, Porcelain; 500ML Each
Mortar and Pestle; Agate, 125ML, Each
Agate Mortar and Pestle, 25 mL (YO-04010-20)
Glass Mortar & Pestle Set, 50ml (YO-34500-51).
More Details
Nylon Filter
Buy Nylon Filter Items Including:
Nylon monofilament filter bag; 10"L, 100 µm (255009-03)
Nylon monofilament filter bag; 10"L, 200 µm (255011-03)
Nylon monofilament filter bag; 10"L, 250 µm (BN-410-250)
Nylon monofilament filter bag; 10"L, 300 µm (255013-03).
More Details
Luer Lock Fittings in Ga
Buy Luer Lock Fittings in Ga Items Including:
Snap luer lock ring, PC, 25/pk (YO-45504-74)
Snap luer lock ring, Nylon, 25/pk (YO-45505-74)
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
Luer fittings, Chrome-plated Luer adapters,male luer lock plug (YO-31507-52).
More Details
Luer Lock Fittings in Georgia
Buy Luer Lock Fittings in Georgia Items Including:
Snap luer lock ring, PC, 25/pk (YO-45504-74)
Snap luer lock ring, Nylon, 25/pk (YO-45505-74)
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
Luer fittings, Chrome-plated Luer adapters,male luer lock plug (YO-31507-52).
More Details
Plastic Bulkhead Fittings Livewell 8
Buy Plastic Bulkhead Fittings Livewell 8 Items Including:
Bulkhead Fitting, PTFE, 3/8" NPT(F) Port X 1-13/16" OD (F-TA03)
Bulkhead Fitting, PVDF, 3/8" NPT(F) Port X 1-13/16" OD (FK-TA03)
Bulkhead Fitting, PVDF, 1-1/4" NPT(F) Port X 2-7/8" OD (FK-TA10).
More Details
Aquarium Water Test Strips
Buy Aquarium Water Test Strips Items Including:
LaMotte Alkalinity Test Strips, 0 to 180 ppm
LaMotte Free Chlorine Test Strips, 0 to 10 ppm
LaMotte Total Chlorine Test Strips, 0 to10 ppm
LaMotte Total Chlorine Test Strips, 0 to 5 ppm
EM Quant Ammonium Test Strips
More Details
Bleach Water Testing Strips
Buy Bleach Water Testing Strips Items Including:
LaMotte Alkalinity Test Strips, 0 to 180 ppm
LaMotte Free Chlorine Test Strips, 0 to 10 ppm
LaMotte Total Chlorine Test Strips, 0 to10 ppm
LaMotte Total Chlorine Test Strips, 0 to 5 ppm
EM Quant Ammonium Test Strips
More Details
70618
results
<<<Previous
19
20
21
22
23
24
25
26
27
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
40
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Stopcocks with Luer Connections to Bleach Water Testing Strips
>
31125329-s-toc_2 31125329stoc2