Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 25
Luer Lock Fittings Moldel
to
Turbidity Calibration Standards
Image
Product Detail
More Detail
Luer Lock Fittings Moldel
Buy Luer Lock Fittings Moldel Items Including:
Snap luer lock ring, PC, 25/pk (YO-45504-74)
Snap luer lock ring, Nylon, 25/pk (YO-45505-74)
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
Luer fittings, Chrome-plated Luer adapters,male luer lock plug (YO-31507-52).
More Details
Luer Lock Fittings Mtll210 Jk1a
Buy Luer Lock Fittings Mtll210 Jk1a Items Including:
Snap luer lock ring, PC, 25/pk (YO-45504-74)
Snap luer lock ring, Nylon, 25/pk (YO-45505-74)
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
316 Stainless Steel fittings, male luer lock x male luer lock (YO-31507-90).
More Details
Plastic Luer Lock Fittings
Buy Plastic Luer Lock Fittings Items Including:
Snap luer lock ring, PC, 25/pk (YO-45504-74)
Snap luer lock ring, Nylon, 25/pk (YO-45505-74)
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
Luer fittings, Chrome-plated Luer adapters,male luer lock plug (YO-31507-52).
More Details
Sealing Luer Lock Fittings Air
Buy Sealing Luer Lock Fittings Air Items Including:
Snap luer lock ring, PC, 25/pk (YO-45504-74)
Snap luer lock ring, Nylon, 25/pk (YO-45505-74)
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
316 Stainless Steel fittings, male luer lock x male luer lock (YO-31507-90).
More Details
Cole-Parmer Manual Vacuum Valves
Buy Cole-Parmer Manual Vacuum Valves Items Including:
Vacuum isolation valve, NW 25 connections.
More Details
Check Valve for Jet Pump
Buy Check Valve for Jet Pump Items Including:
Nylon Miniature Check Valve, 1/8" hose barb, fluorosilicone diaphragm (YO-98553-00)
Nylon Miniature Check Valve, 3/16" hose barb, fluorosilicone diaphragm (YO-98553-01)
PVDF Miniature Check Valve, 3/16" hose barb, Viton diaphragm.
More Details
5 Ml Volumetric Flask Tolerance
Buy 5 Ml Volumetric Flask Tolerance Items Including:
PYREX Brand 5640 Volumetric Flask; 100 mL, pack of 6
PYREX Brand 5640 Volumetric Flask; 200 mL, pack of 6
PYREX Brand 5640 Volumetric Flask; 1000 mL, pack of 1
PYREX Brand 5640 Volumetric Flask; 2000 mL, pack of 1.
More Details
Flare to 3 8 Compression Fittings
Buy Flare to 3 8 Compression Fittings Items Including:
Flare compression pipe adapter, 1/4" x 3/8", PTFE, Each (YO-30490-22)
Flare compression pipe adapter, 3/8" x 1/8", PTFE, Each (YO-30490-26)
Flare compression pipe adapter, 3/8" x 1/4", PTFE, Each (YO-30490-28).
More Details
Mbp Art Aerosol-Resistant Pipette Tips
Buy Mbp Art Aerosol-Resistant Pipette Tips Items Including:
Tip Art .1-10µL Sterile 960/pk (2139-05)
Tip Art 5-200µL Sterile 960/pk (2160P)
Tip Art 1-300µL Sterile 960/pk (2070)
Tip Art 100-1000µL Sterile 768/pk (2279)
Tip Art 100-1000µL Sterile 800/pk (2079E)
More Details
Respirator Cartridge
Buy Respirator Cartridge Items Including:
S-series Respirator Cartridges, Organic Vapor, with P100 filter, 4/box
S-series Respirator Cartridges, Acid Gases, 6/box
S-series Respirator Cartridges, Acid Gases, with P100 filter, 4/box
S-series Respirator Cartridges, Multi-Contaminant, 6/box.
More Details
Electronic Metering Pumps
Buy Electronic Metering Pumps Items Including:
Feed-Verification Diaphragm Metering Pump, 5.4 GPH, PVC head, EPDM dia
Programmable Remote Control Solenoid-driven Diaphragm Metering Pump wi (EHE30E1-PC)
Programmable Remote Control Solenoid-driven Diaphragm Metering Pump wi (EHE30E2-PC).
More Details
Allsafe Safety Glasses
Buy Allsafe Safety Glasses Items Including:
Allsafe SMC™ Nemesis™ Safety Glasses, Black frame, Clear lens
Allsafe MonoGoggle, Regular, Clear Lens
Allsafe MonoGoggle, Large, Clear Lens
Allsafe MonoGoggle XTR™, Blue Frame, Clear Lens
Allsafe Shield/MonoGoggle XTR™, Blue Frame, Clear Lens.
More Details
Stainless Steel Barbed Tee
Buy Stainless Steel Barbed Tee Items Including:
Tee, Stainless Steel, 1/8", Each (YO-30620-25)
Tee, Stainless Steel, 3/16", Each (YO-30620-26)
Tee, Stainless Steel, 5/16", Each (YO-30620-28)
Male Branch Tee, 316 SS, 10-32 UNF(M) x 1/16" hose barb (YO-31209-50).
More Details
Graduated Cylinders 2 Liter Poly
Buy Graduated Cylinders 2 Liter Poly Items Including:
Cole-Parmer Graduated Cylinder, PP 10mL, 2/pk (CP0010)
Cole-Parmer Graduated Cylinder, PP 25mL, 2/pk (CP0025)
Cole-Parmer Graduated Cylinder, PP 50mL, 2/pk (CP0050)
Corning Graduated Cylinder 10ml PP, 2/pk
Corning Graduated Cylinder 50ml PP, 2/pk.
More Details
Cole-Parmer Pvdf Sanitary Clamp Tee Fittings
Buy Cole-Parmer Pvdf Sanitary Clamp Tee Fittings Items Including:
PVDF Sanitary Tee, 3/4" Mini Clamp Size (YO-31809-61)
PVDF Sanitary Tee, 1" Ladish Clamp Size (YO-31809-62)
PVDF Sanitary Tee, 1 1/2" Clamp Size (YO-31809-63)
PVDF Sanitary Tee, 2 1/2" Clamp Size (YO-31809-65).
More Details
Food Grade Stainless Steel Tubing
Buy Food Grade Stainless Steel Tubing Items Including:
316 Stainless Steel Tubing; dimensions, 1/16" ID, 1/8" OD, 1/32" wall; (YO-03300-05)
304 Stainless Steel Tubing, 1/8" OD, 0.085" ID, 50ft roll (YO-34680-50)
Stainless steel tube set for electric and air motors, 27"L.
More Details
Ohaus Scales
Buy Ohaus Scales Items Including:
Ohaus Floor Ramp, 60" Wide for VN Floor Scale (YO-11600-22)
Ohaus Floor Ramp, 48" Wide for 10k VX Floor Scale
Ohaus Floor Ramp, 60" Wide for 10k VX Floor Scale
Ohaus Fixed location foot mounting plate kit
Ohaus Feet for VX (4 pack).
More Details
Scienceware Churn Sample Splitters
Buy Scienceware Churn Sample Splitters Items Including:
Scienceware 4 liter Churn Sample Splitter
Scienceware 8 liter Churn Sample Splitter.
More Details
Large Industrial Funnels
Buy Large Industrial Funnels Items Including:
Large industrial-size low-density polyethylene funnel, 1.5 L (YO-06121-10)
Large industrial-size low-density polyethylene funnel, 4 L (YO-06121-20)
Large industrial-size polypropylene funnel, 4.5 L (YO-06121-32).
More Details
Precision Stainless Steel Tubing
Buy Precision Stainless Steel Tubing Items Including:
316 Stainless Steel Tubing; dimensions, 1/16" ID, 1/8" OD, 1/32" wall; (YO-03300-05)
304 Stainless Steel Tubing, 1/8" OD, 0.085" ID, 50ft roll (YO-34680-50)
Stainless steel tube set for electric and air motors, 27"L.
More Details
25 Ml Graduated Cylinder Plastic
Buy 25 Ml Graduated Cylinder Plastic Items Including:
Thermo Scientific Nalgene graduated polymethylpentene cylinder, 25 mL
Thermo Scientific Nalgene graduated polypropylene cylinder, 25 mL
Cole-Parmer Graduated Cylinder, PP 10mL, 24/pk (CP0010)
Cole-Parmer Graduated Cylinder, PP 25mL, 12/pk (CP0025).
More Details
Cole-Parmer Solenoid Valves
Buy Cole-Parmer Solenoid Valves Items Including:
Mounting Flange for Cole-Parmer Solenoid Valves
Cole-Parmer Solenoid Pinch Valve; for use with L/S 14 tubing, normally (PK-1605NC-12V)
Cole-Parmer Solenoid Pinch Valve; for use with L/S 14 tubing, normally (PK-1605NC-24V).
More Details
Nalgene 1 Gallon Hdpe Bottles
Buy Nalgene 1 Gallon Hdpe Bottles Items Including:
Thermo Scientific Nalgene Wide-Mouth Economy Bottle, HDPE, 1000 mL, 6/ (YO-06035-51)
Thermo Scientific Nalgene Wide-Mouth Economy Bottle, HDPE, 1000 mL, 24 (YO-06035-53)
Thermo Scientific Nalgene heavy-duty polypropylene bottle, 1 L.
More Details
Pyrex Brand 9825 Culture Tubes
Buy Pyrex Brand 9825 Culture Tubes Items Including:
Pyrex Brand 9825 culture tube with screw cap; 11 mL, pack of 144
Pyrex Brand 9825 culture tube with screw cap; 15 mL, pack of 144
Pyrex Brand 9825 culture tube with screw cap; 20 mL, pack of 144.
More Details
Large Stainless Steel Funnel with Handle
Buy Large Stainless Steel Funnel with Handle Items Including:
Stainless steel funnel, 2 3/8 oz (YO-07263-00)
Stainless steel funnel, 13 oz (YO-07265-00)
Stainless steel funnel, 22 1/4 oz (YO-07266-00)
Top Cover with Handle for 8" Stainless Steel Sieve
Stainless steel spatula with hardwood handle (2215).
More Details
Round Rubber Tubing 1 2
Buy Round Rubber Tubing 1 2 Items Including:
Black Rubber Tubing, 1/2" ID x 3/4" OD, 25 ft (YO-96142-13)
Black Rubber Vacuum Tubing, 1/2" ID x 1" OD, 25ft
(YO-96143-10)
Black Rubber Vacuum Tubing, 1/2" ID x 1" OD, 25ft
(YO-96143-11)
More Details
Microflex Disposable Latex Gloves
Buy Microflex Disposable Latex Gloves Items Including:
Microflex Synetron Latex Gloves; size, small
Microflex Synetron Latex Gloves; size, medium
Microflex Color Touch Latex Gloves; green; size, medium
Microflex Diamond Grip™ Latex Gloves; size, small
Microflex Diamond Grip™ Latex Gloves; size, large.
More Details
Thermo Scientific Thermolyne Benchtop Muffle Furnaces
Buy Thermo Scientific Thermolyne Benchtop Muffle Furnaces Items Including:
Thermo Scientific Thermolyne Benchtop Muffle Furnace with 120 cu in ch (F47910-33)
Thermo Scientific Thermolyne Benchtop Muffle Furnace with 350 cu in ch (F48015-60)
Thermo Scientific Thermolyne Benchtop Muffle Furnace with 120 cu in ch (F47925-80).
More Details
25 Ml Erlenmeyer Flask
Buy 25 Ml Erlenmeyer Flask Items Including:
Universal 25-ml Erlenmeyer flask clamp
25 ml Erlenmeyer Flask Clip
PYREX Brand 4980 flask; 25 mL, pack of 12
KIMAX flask, 25 mL, 48/CS
PYREX VISTA™ narrow mouth flask, 25 mL, 24/CS
More Details
Turbidity Calibration Standards
Buy Turbidity Calibration Standards Items Including:
Hach StablCal 0.1 NTU Turbidity Standard, 500-mL
Hach StablCal 200 NTU Turbidity Standard, 500-mL
Hach StablCal 1000 NTU Turbidity Standard, 500-mL
HF scientific Calibration Kit for MICROTOL,3 Turbidimeter
Turbidity Standard, 10.0 NTU, 1-L bottle.
More Details
70618
results
<<<Previous
21
22
23
24
25
26
27
28
29
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Luer Lock Fittings Moldel to Turbidity Calibration Standards
>
31125329-s-toc_2 31125329stoc2