Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 26
250 Ml Erlenmeyer Flask
to
Thermo Scientific Nalgene Unitary Wash Bottles
Image
Product Detail
More Detail
250 Ml Erlenmeyer Flask
Buy 250 Ml Erlenmeyer Flask Items Including:
Corning 250 mL PC Flask w/vented cap, 50/CS
Thermo Scientific Low Cost Orbital Shaker for 250 mL Erlenmeyer flasks (2346Q)
Thermo Scientific Low Cost Orbital Shaker for 250 mL Erlenmeyer flasks (2346-1CEQ)
PYREX Brand 4995 flask; 250 mL, case of 6.
More Details
Atc Probe for Corning Ph Meter
Buy Atc Probe for Corning Ph Meter Items Including:
ATC Probe (477968)
Oakton Acorn pH 5 meter with electrode, and ATC probe (pH 5)
Oakton® Acorn™ pH 5 meter with ATC probe (pH 5)
Oakton® Acorn™ pH 6 meter with ATC probe (pH 6)
Oakton pH 310 meter with single-junction pH/ATC probe with NIST-tracea.
More Details
Drinking Water Test Strips Copper
Buy Drinking Water Test Strips Copper Items Including:
Watersafe Drinking Water Test Kit
LaMotte Free Chlorine Test Strips, 0 to 10 ppm
LaMotte Total Chlorine Test Strips, 0 to10 ppm
LaMotte Total Chlorine Test Strips, 0 to 5 ppm
LaMotte Hardness Test Strips, 0 to 180 ppm
More Details
Pyrex Erlenmeyer Flask
Buy Pyrex Erlenmeyer Flask Items Including:
Pyrex Glass Flask Starter Set
Pyrex 5020 Flasks, 25mL, 6/pk
Pyrex 5020 Flasks, 125mL, 6/pk
Pyrex 5020 Flasks, 250mL, 6/pk
Pyrex 5020 Flasks, 500mL, 1/pk
More Details
3l Pyrex Erlenmeyer Flasks
Buy 3l Pyrex Erlenmeyer Flasks Items Including:
Pyrex 5020 Flasks, 25mL, 6/pk
Pyrex 5020 Flasks, 50mL, 6/pk
Pyrex 5020 Flasks, 125mL, 6/pk
Pyrex 5020 Flasks, 250mL, 6/pk
Pyrex 5020 Flasks, 500mL, 1/pk
More Details
Ysi Thermistor Probes and Accessories
Buy Ysi Thermistor Probes and Accessories Items Including:
700 Series Interchangeable Thermistor General Purpose Probe.
More Details
Microflex Colortouch Pink Latex Glove
Buy Microflex Colortouch Pink Latex Glove Items Including:
Microflex ColorTouch Pink Powder-Free Gloves, Small
Microflex ColorTouch Pink Powder-Free Gloves, Medium
Microflex ColorTouch Pink Powder-Free Gloves, Large.
More Details
Digital Thermometer
Buy Digital Thermometer Items Including:
Taylor 9847N Pen-Style Waterproof Digital Thermometer with Anti-Microb
Cooper-Atkins DFP450W Waterproof digital thermometer with reduced tip
Wd-03316-80:Barometer W Digital Thermometer (YO-03316-80)
Oakton Thermohygrometer with Digital Thermometer.
More Details
Standard Duty Dry Vacuum Piston Pumps
Buy Standard Duty Dry Vacuum Piston Pumps Items Including:
Standard Duty Dry Vacuum Piston Pumps, 22 L/min, 115 VAC
Standard Duty Dry Vacuum Piston Pumps, 19 L/min, 230 VAC
Standard Duty Dry Vacuum Piston Pumps, 34 L/min, 115 VAC
Standard Duty Dry Vacuum Piston Pumps, 201 L/min, 115 VAC.
More Details
Delmhorst Moisture Meters
Buy Delmhorst Moisture Meters Items Including:
Delmhorst J-Lite Wood Moisture Meter 6-30%
Delmhorst J-4 Wood Moisture Analog Meter 6 to 30%
Replacement pins - 2" insulated pins
Replacement PINS - 7IN
Replacement PINS - 1-1/2in insulated pins
More Details
Adapters Hose to Threaded Male Pipe
Buy Adapters Hose to Threaded Male Pipe Items Including:
Barbed fittings, NPT male pipe adapter, Natural PP, 1/8" NPT x 1/4" ID (YO-30610-36)
Barbed fittings, NPT male pipe adapter, Natural PP, 1/8" NPT x 3/8" ID (YO-30610-37).
More Details
Mechanical Oil Pressure Gauges
Buy Mechanical Oil Pressure Gauges Items Including:
Cole-parmer Industrial Pressure Gauge, 30" Hg To 60 PSI; 1/4" NPT(M)
Cole-parmer Industrial Pressure Gauge, 30" Hg To 30 PSI; 1/4" NPT(M)
Cole-parmer Industrial Pressure Gauge, 30" Hg To 200 PSI; 1/4" NPT(M).
More Details
Cole-Parmer Polypropylene Sanitary Clamp Tee Fittings
Buy Cole-Parmer Polypropylene Sanitary Clamp Tee Fittings Items Including:
Polypropylene Sanitary Tee, 3/4" Mini Clamp Size (YO-31806-61)
Polypropylene Sanitary Tee, 1" Ladish Clamp Size (YO-31806-62)
Polypropylene Sanitary Tee, 1 1/2" Clamp Size (YO-31806-63).
More Details
Thermo Scientific Nunc Centrifuge Tubes
Buy Thermo Scientific Nunc Centrifuge Tubes Items Including:
Thermo Scientific Nunc™ EZFlip™ Conical-Bottom Sterile 15 (YO-17411-30)
Thermo Scientific Nunc™ EZFlip™ Conical-Bottom Sterile 15 (YO-17411-32)
Thermo Scientific Nunc Conical Centrifuge Tubes 500/CS, RACKED 15mL.
More Details
To 5 16 Barb Reducer
Buy To 5 16 Barb Reducer Items Including:
Barbed fittings, Reducing Connector, Natural PP, 5/8" x 5/16" ID, 3/16 (YO-30610-14)
Reducing Connector, Polyethylene, 5/8" x 5/16", 10 PACK (YO-30621-98)
Reducing Connector, Nylon, 5/8" x 5/16", 10 PACK (YO-30621-99).
More Details
Luxo Illuminated Magnifier Accessories
Buy Luxo Illuminated Magnifier Accessories Items Including:
10-Diopter Swing-away Auxiliary Lens.
More Details
Thermo Orion Ph Meters
Buy Thermo Orion Ph Meters Items Including:
Thermo Scientific Orion pH electrode, epoxy body, sealed, double-junct
Thermo Scientific Orion 3-Star handheld pH meter
Thermo Scientific Orion Star LogR pH Meter Only
Thermo Scientific Orion pH/ATC Triode, epoxy body, sealed, single-junc (9109WP).
More Details
Cole-Parmer Compact Electromagnetic Sieve Shaker
Buy Cole-Parmer Compact Electromagnetic Sieve Shaker Items Including:
Compact Electromagnetic Sieve Shaker; 115 VAC, 60 Hz.
More Details
Wika Stainless Steel Digital Gauges
Buy Wika Stainless Steel Digital Gauges Items Including:
WIKA Stainless Steel Digital Gauge ~ 30" Hg to 29 psi, 1/4" NPT(M)
WIKA Stainless Steel Digital Gauge ~ 30" Hg to 72 psi, 1/4" NPT(M)
WIKA Stainless Steel Digital Gauge ~ 30" Hg to 145 psi, 1/4" NPT(M).
More Details
Corning Electrode
Buy Corning Electrode Items Including:
Replacement three-in-one pH electrode, gel-filled junction; 6-ft cable
Replaceable electrode; glass body, refillable, no ATC
pH electrode refillable single-junction for Corning meters (YO-27507-28)
ATC Probe (477968)
Single-junction (YO-27507-20).
More Details
Touch-N-Tuff Disposable Nitrile Gloves
Buy Touch-N-Tuff Disposable Nitrile Gloves Items Including:
Ansell Touch-N-Tuff™ Nitrile Gloves, Powder-free, Small
Ansell Touch-N-Tuff™ Nitrile Gloves, Powder-free, Medium
Ansell Touch-N-Tuff™ Nitrile Gloves, Powder-free, Large
Ansel Touch-N-Tuff™ Nitrile Gloves, Powdered, Extra-large.
More Details
Infrared Thermometer Calibration Service
Buy Infrared Thermometer Calibration Service Items Including:
NIST-Traceable Calibration; Thermometer Probe used with IR Thermometer (YO-17004-10).
More Details
Amazon Well Water Test Strips
Buy Amazon Well Water Test Strips Items Including:
Watersafe Well Water Test Kit
LaMotte Alkalinity Test Strips, 0 to 180 ppm
LaMotte Free Chlorine Test Strips, 0 to 10 ppm
LaMotte Total Chlorine Test Strips, 0 to10 ppm
LaMotte Total Chlorine Test Strips, 0 to 5 ppm
More Details
Medical Equipment Calibration Services
Buy Medical Equipment Calibration Services Items Including:
Innocal NIST Calibration for Refrigerators & Freezers
Innocal NIST Calibration for Incubators.
More Details
Kimax Glassware 1000ml Media Bottles
Buy Kimax Glassware 1000ml Media Bottles Items Including:
1000mL Media Storage Bottles (14395-1000)
500 mL Kimax Amber Media Bottles
1000 mL Kimax Amber Media Bottles
Replacement cap for Kimax media bottles, PP, blue, 10/cs
100mL Media Storage Bottles (14395-100)
More Details
Double Stir Bars
Buy Double Stir Bars Items Including:
Double Stir Bar, 1/2"H x 1/2"dia (37126-9002)
Double Stir Bar, 3/4"H x 5/8"dia (37126-9003).
More Details
Cole-Parmer Natural Polypropylene Barbed Reducing Connectors
Buy Cole-Parmer Natural Polypropylene Barbed Reducing Connectors Items Including:
Barbed fittings, Reducing Connector, Natural PP,5/16" x 1/4" ID, 1/8", (YO-30610-07)
Barbed fittings, Reducing Connector, Natural PP, 3/8" x 1/4" ID, 1/8", (YO-30610-08).
More Details
Cole-Parmer Round Jugs, Hdpe
Buy Cole-Parmer Round Jugs, Hdpe Items Including:
Cole-Parmer Round 1/2 Gallon Jug, HDPE, 24/pk.
More Details
I-Lab Handheld Analyzing Spectrometers
Buy I-Lab Handheld Analyzing Spectrometers Items Including:
Spectrum Programmable Software for i-LAB
Round Sample Vials for i-LAB, 25-mm Diameter; Pack of 6
i-LAB Surface Reader Adaptor
i-LAB 25-mm Round Vial Adaptor.
More Details
Thermo Scientific Nalgene Unitary Wash Bottles
Buy Thermo Scientific Nalgene Unitary Wash Bottles Items Including:
Thermo Scientific Nalgene wide-mouth Unitary wash bottle, 125 mL
Thermo Scientific Nalgene wide-mouth Unitary wash bottle, 250 mL
Thermo Scientific Nalgene wide-mouth Unitary wash bottle, 500 mL.
More Details
70618
results
<<<Previous
22
23
24
25
26
27
28
29
30
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
250 Ml Erlenmeyer Flask to Thermo Scientific Nalgene Unitary Wash Bottles
>
31125329-s-toc_2 31125329stoc2