Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 27

Oilless Air Compressor   to   Pyrex Vacuum Desiccators

Image
  Product Detail
More Detail

Oilless Air Compressor, Piston Compressor Pumps

Buy Oilless Air Compressor, Piston Compressor Pumps Items Including:
  • Oilless Air Compressor, Piston compressor pump, 3.5 cfm, 115/230 VAC (4HCC-10-M450X)
  • Oilless Air Compressor, Piston compressor pump, 9.1 cfm, 220/380 VAC (7HDD-10-M853)
  • Oilless Air Compressor, Piston compressor pump, 11 cfm, 230 VAC (8HDM-10-M850X).

Air Compressor Fittings

Buy Air Compressor Fittings Items Including:
  • Air Compressor hose, 1/4 in x 25' PU braid air hose with swivel fittin (AA1061-blue)
  • Air Compressor hose, 1/4 in x 50' PU braid air hose with swivel fittin (AA1062-blue).

Myron L Conductivity Calibration Solutions

Buy Myron L Conductivity Calibration Solutions Items Including:
  • MYRON L conductivity calibration standard, 23.8 µS
  • MYRON L conductivity calibration standard, 46.7 µS
  • MYRON L conductivity calibration standard, 2060 µS
  • MYRON L conductivity calibration standard, 16,630 µS.

Kimble Separatory Funnels

Buy Kimble Separatory Funnels Items Including:
  • Kimble Separatory Funnel, PTFE Stopcock, 1000 mL 2/cs (YO-34505-50)
  • Kimble Separatory Funnel, PTFE Stopcock, 2000 mL 2/cs (YO-34505-52)
  • Kimble Separatory Funnel, 250mL, 4/cs (YO-34507-14)
  • Kimcote Separatory Funnel With Glass Stopper, PTFE, 2000 ml.

Compression Tee Fitting 1 8

Buy Compression Tee Fitting 1 8 Items Including:
  • Compression fittings, Union Tees, Kynar, 1/8" OD, 1-1/8", 3/4", 3/32"; (YO-30805-21)
  • Union Tee, Brass, 1/8" x 1/8", Each (YO-31412-91)
  • Tee union, 1/8", Brass, each (YO-31412-17)
  • Copper Tee Fitting 1 1/4in Pipe Id.

Food Grade Plastic Check Valves

Buy Food Grade Plastic Check Valves Items Including:
  • Hayward True Union Ball Check Valve, 1/2" socket and NPT(F) connectors
  • Hayward True Union Ball Check Valve, 3/4" socket and NPT(F) connectors
  • Hayward True Union Ball Check Valve, 1-1/2" socket and NPT(F) connecto.

Inch Plastic Check Valves

Buy Inch Plastic Check Valves Items Including:
  • Hayward True Union Ball Check Valve, 1/2" socket and NPT(F) connectors
  • Hayward True Union Ball Check Valve, 3/4" socket and NPT(F) connectors
  • Hayward True Union Ball Check Valve, 1-1/2" socket and NPT(F) connecto.

Inline Plastic Check Valve

Buy Inline Plastic Check Valve Items Including:
  • Hayward True Union Ball Check Valve, 1/2" socket and NPT(F) connectors
  • Hayward True Union Ball Check Valve, 3/4" socket and NPT(F) connectors
  • Hayward True Union Ball Check Valve, 1-1/2" socket and NPT(F) connecto.

Plastic Coolant Check Valves

Buy Plastic Coolant Check Valves Items Including:
  • Hayward True Union Ball Check Valve, 1/2" socket and NPT(F) connectors
  • Hayward True Union Ball Check Valve, 3/4" socket and NPT(F) connectors
  • Hayward True Union Ball Check Valve, 1-1/2" socket and NPT(F) connecto.

A&D Weighing Phoenix Gh Analytical Balances

Buy A&D Weighing Phoenix Gh Analytical Balances Items Including:
  • A&D Phoenix Series Analytical Balance 120g x 0.1mgwith Internal Calibr
  • A&D Phoenix Series Analytical Balance 220g x 0.1mg with Internal Calib
  • A&D Phoenix Series Analytical Balance 320g x 0.1mg with Internal Calib.

Thermo Scientific Nanodrop Lite Spectrophotometer

Buy Thermo Scientific Nanodrop Lite Spectrophotometer Items Including:
  • Thermo Scientific NanoDrop Lite Spectrophotometer with Printer, 120V
  • Thermo Scientific NanoDrop Lite Spectrophotometer 120V
  • Thermo Scientific NanoDrop Lite Printer Paper Refill 2/pk.

Industrial Mixers

Buy Industrial Mixers Items Including:
  • Industrial Mixer Lift Stand, Manual (YO-04600-10)
  • Lightnin small batch, fixed-speed mixer with 1/20-hp explosion-proof m
  • Lightnin small batch fixed-speed mixer with 1/8-hp motor, 115 VAC
  • Lightnin heavy-duty, gear-drive mixer, TENV (YO-50310-00).

Coorstek High Alumina Crucibles

Buy Coorstek High Alumina Crucibles Items Including:
  • CoorsTek High-Form Crucible, High Alumina; 10ml
  • CoorsTek High-Form Crucible, High Alumina; 20ml
  • CoorsTek High-Form Crucible, High Alumina; 50ml
  • CoorsTek High-Form Crucible, High Alumina; 100ml
  • CoorsTek High-Form Crucible Cover, High Alumina, 10ML.

Thermo Scientific Nalgene Trays

Buy Thermo Scientific Nalgene Trays Items Including:
  • Thermo Scientific Nalgene polypropylene autoclaving tray, 1 L
  • Thermo Scientific Nalgene polypropylene autoclaving tray, 2 L
  • Thermo Scientific Nalgene polypropylene autoclaving tray, 3 L
  • Thermo Scientific Nalgene polypropylene autoclaving tray, 5 L.

Lab-Tek Chamber Slide Systems

Buy Lab-Tek Chamber Slide Systems Items Including:
  • Thermo Scientific Nunc Lab-Tek™ Chamber Slide™ System; material, (YO-01838-20)
  • Thermo Scientific Nunc Lab-Tek™ Chamber Slide™ System; material, (YO-01838-21)
  • Thermo Scientific Nunc Lab-Tek™ Chamber Slide™ System; material, (YO-01838-22).

Rotary Vane Vacuum Pump Oil

Buy Rotary Vane Vacuum Pump Oil Items Including:
  • Vacuubrandreg Rotary Vane Vacuum Pump, 1.6 cfm, 1.5 X 10-3/sup (698123)
  • Vacuubrandreg Rotary Vane Vacuum Pump, 1.6 cfm, 1.5 X 10-3/sup (698120)
  • Vacuubrandreg Rotary Vane Vacuum Pump, 4.0 cfm, 1.5 X 10-3/sup (698133).

Brass Barb Fittings

Buy Brass Barb Fittings Items Including:
  • Barbed fittings, Metal quick-disconnect fittings, coupling bodies,valv (LCD17005)
  • Barbed fittings, Metal quick-disconnect fittings, coupling bodies,valv (LCD17006)
  • Barbed fittings, metal quick-disconnect fittings, inserts,valved fitti (LCD22004).

Stainless Steel Sieves with Brackets

Buy Stainless Steel Sieves with Brackets Items Including:
  • Receiving Pan with Fitted Rim for Nesting 12" Stainless Steel Sieves, (YO-59996-92)
  • Receiving Pan with Fitted Rim for Nesting 12" Stainless Steel Sieves, (YO-59996-93)
  • Receiving Pan for 8" Diameter Stainless Steel Full Height Sieves.

Reducer to 1 4 Compression Fitting

Buy Reducer to 1 4 Compression Fitting Items Including:
  • Compression reducing union, Pharmafluor™ PTFE, 1/4" OD x 1/8" OD (YO-06379-11)
  • Reducing compression union, polypropylene, 5/16 x 1/4" OD, 10 per pack (YO-06381-70)
  • Reducing Union, 5/16" x 1/4", Nylon, 10 Pack (YO-06382-17).

Corning Ph Electrode

Buy Corning Ph Electrode Items Including:
  • Replacement three-in-one pH electrode, gel-filled junction; 6-ft cable
  • pH electrode refillable single-junction for Corning meters (YO-27507-28)
  • Replaceable electrode; glass body, refillable, no ATC
  • ATC Probe (477968)
  • Single-junction (YO-27507-20).

Wheaton Glass Bottles W216916

Buy Wheaton Glass Bottles W216916 Items Including:
  • WHEATON BOD Bottle, 300 mL, glass, unnumbered, penny head stopper, 24/
  • WHEATON BOD Bottle, 300 mL, glass, unnumbered, robotic stopper, 24/cas
  • Clear glass dropping bottle, 100 mL
  • Pack of 6
  • (W211735)

Serum Vials

Buy Serum Vials Items Including:
  • Scienceware serum vial rack for 10- to 13-mm vials
  • Scienceware serum vial rack for 13- to 16-mm vials
  • Scienceware serum vial rack for 16- to 20-mm vials
  • Clear Serum Tubing Vial, 2 mL, 144/cs (223683)
  • Clear Serum Tubing Vial, 3 mL, 144/cs (223684).

3-Way Solenoid Pinch Valves

Buy 3-Way Solenoid Pinch Valves Items Including:
  • Three-way solenoid pinch valve; 12 VDC, 1/8" ID x 1/4" OD tubing (100P3MP-12-05)
  • Solenoid-Operated Two-Way Pinch Valve; Normally-Closed, 13 mm Tube OD, (EPK-1005-NC 24V N CLOSED).

Tough-Tags Microtube Labels

Buy Tough-Tags Microtube Labels Items Including:
  • Small Tough-Tags™ label, clear
  • Small Tough-Tags™ label, white
  • Small Tough-Tags™ label, red
  • Small Tough-Tags™ label, yellow
  • Small Tough-Tags™ label, green

1 Inch Sanitary to Luer Fitting

Buy 1 Inch Sanitary to Luer Fitting Items Including:
  • Adapter, Kynar, 1/2" mini to 1/2" to female luer lock
  • Luer fittings, Chrome-plated Luer adapters,male luer lock to 1/8" to 3 (YO-31507-61)
  • Luer fittings, Chrome-plated Luer adapters,female luer to 1/8" to 3/16 (YO-31507-63).

Npt X 3 8 Barb Tee

Buy Npt X 3 8 Barb Tee Items Including:
  • Barbed fittings, NPT male branch tees, Kynar, 1/8" NPT x 3/8" ID, 1/4" (YO-30704-62)
  • Barbed fittings, NPT male branch tees, Kynar, 1/4" NPT x 3/8" ID, 1/4" (YO-30704-64).

John Guest Valves

Buy John Guest Valves Items Including:
  • John Guest Speedfit Ball Valve, straight, 1/4" OD speedfit x 1/4" NPT(
  • Ball valve, 2-way right angled, 3/8" NPT(F) x 3/8" John Guest (PVC-657-6F6CJG-B)
  • Ball Valve, 2-way, 3/8" John Guest, PVC with Buna N seals (YO-01379-13).

Kynar Tubing

Buy Kynar Tubing Items Including:
  • Kynar tubing, 11/64"ID x 1/4"OD, 25 ft/pack
  • Kynar® tube set for 07075-series motors, 40"L (16520-3400)
  • Tubing Adapter; Kynar, 1/2" mini to 1/4" NPT (M)
  • Tubing Adapter; Kynar, 1/2" mini to 3/8" NPT (M).

Miniature Hose Barbs

Buy Miniature Hose Barbs Items Including:
  • Nylon Miniature Check Valve, 1/8" hose barb, fluorosilicone diaphragm (YO-98553-00)
  • Nylon Miniature Check Valve, 3/16" hose barb, fluorosilicone diaphragm (YO-98553-01)
  • PVDF Miniature Check Valve, 3/16" hose barb, Viton diaphragm.

Pyrex Vacuum Desiccators

Buy Pyrex Vacuum Desiccators Items Including:
  • Pyrex Vacuum Desiccator, 5.8L
  • Pyrex Vacuum Desiccator, 2.4L
  • Pyrex Vacuum Desiccator, 10.5L
  • Pyrex Desiccator, 10.5L
  • Pyrex Desiccator, 2.4L.



Oilless Air Compressor to Pyrex Vacuum Desiccators

>