Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 29
Oakton Ph Buffer Coa
to
Rubber Stopper Size Chart
Image
Product Detail
More Detail
Oakton Ph Buffer Coa
Buy Oakton Ph Buffer Coa Items Including:
Oakton pH buffer pouches; pH 4.01, 20/pk
Oakton pH buffer pouches; pH 7.00, 20/pk
Oakton pH buffer pouches; pH 10.00, 20/pk
Oakton 4.01 pH buffer, 500-mL
Oakton 1.68 pH buffer, 500-mL
More Details
Oakton Ph Buffer Kit
Buy Oakton Ph Buffer Kit Items Including:
Oakton pH buffer pouches; pH 4.01, 20/pk
Oakton pH buffer pouches; pH 7.00, 20/pk
Oakton pH buffer pouches; pH 10.00, 20/pk
Oakton 4.01 pH buffer, 500-mL
Oakton 1.68 pH buffer, 500-mL
More Details
Oakton Ph Buffer Msds
Buy Oakton Ph Buffer Msds Items Including:
Oakton pH buffer pouches; pH 4.01, 20/pk
Oakton pH buffer pouches; pH 7.00, 20/pk
Oakton pH buffer pouches; pH 10.00, 20/pk
Oakton 4.01 pH buffer, 500-mL
Oakton 1.68 pH buffer, 500-mL
More Details
Oakton Ph Buffer Scale
Buy Oakton Ph Buffer Scale Items Including:
Oakton pH buffer pouches; pH 4.01, 20/pk
Oakton pH buffer pouches; pH 7.00, 20/pk
Oakton pH buffer pouches; pH 10.00, 20/pk
Oakton 4.01 pH buffer, 500-mL
Oakton 1.68 pH buffer, 500-mL
More Details
Cole-Parmer Combs for Gel Electrophoresis Systems
Buy Cole-Parmer Combs for Gel Electrophoresis Systems Items Including:
Comb for Cole-Parmer Horizontal Mini-Gel System Model 28559-00; 8/10 W (YO-28559-50)
Comb for Cole-Parmer Horizontal Mini-Gel System Model 28559-00; 8/10 W (YO-28559-52).
More Details
Sample Line with Male Luer Fitting
Buy Sample Line with Male Luer Fitting Items Including:
Adapters with 1/4-28 UNF thread; male luer fitting; ETFE (YO-06471-90)
Adapters with caps; male luer fitting; Tefzel
Male luer x male luer adapter, PC, 25/pk (YO-45504-72)
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55).
More Details
Bausch & Lomb Handheld Magnifiers
Buy Bausch & Lomb Handheld Magnifiers Items Including:
Bausch & Lomb Handheld LED Magnifier, 2" x 4", 2x magnification
Bausch & Lomb Handheld LED Magnifier, 3-1/2" diameter, 2x magnificatio
Measuring Scales;" scale, 0.75" in 0.005" increments (81-34-37).
More Details
Texwipe Lint Free Cleanroom Wipes
Buy Texwipe Lint Free Cleanroom Wipes Items Including:
Texwipe QuanSat Prewetted Cleanroom Wipes (IPA 100%), 9" x 9", 50/pack
Texwipe Vectra Alpha Nu Low-Particulate Cleanroom Wipes, 9" x 9", 150/
Texwipe AlphaWipe Low-particulate wipes; 9" x 9", bulk packed, 150/bg.
More Details
Cole-Parmer Low-Pressure-Drop Liquid Flow Switches
Buy Cole-Parmer Low-Pressure-Drop Liquid Flow Switches Items Including:
Normally open flow switch for liquids; 0.5 GPM (YO-32776-00)
Normally open flow switch for liquids; 1.0 GPM (YO-32776-01)
Normally open flow switch for liquids; 2.0 GPM (YO-32776-02)
Normally open flow switch for liquids; 3.0 GPM (YO-32776-03).
More Details
Thermo Scientific Finnpipette Multichannel Pipettors
Buy Thermo Scientific Finnpipette Multichannel Pipettors Items Including:
Thermo Scientific Finnpipette Novus Multichannel 8 Channel, 5 to 50 m
Thermo Scientific Finnpipette Novus Multichannel 8 Channel, 100 to 120
Thermo Scientific Finnpipette Novus Multichannel 12 Channel, 30 to 300.
More Details
Moyno Pumps
Buy Moyno Pumps Items Including:
Moyno progressing cavity pump, phenolic housing and rotor/nitrile stat (20551)
Moyno progressing cavity pump, 316 stainless steel housing and rotor/n (3910000251)
Moyno progressing cavity pump, phenolic housing and rotor/nitrile stat (3910000252).
More Details
Pyrex Brand 3140 Dishes
Buy Pyrex Brand 3140 Dishes Items Including:
Pyrex Brand 3140 dish; 100 x 50 mm, pack of 6
Pyrex Brand 3140 dish; 125 x 65 mm, pack of 4
Pyrex Brand 3140 dish; 150 x 75 mm, pack of 4
Pyrex Brand 3140 dish; 170 x 90 mm, pack of 2
Pyrex Brand 3140 dish; 190 x 100 mm, pack of 2.
More Details
Orion Ph Electrode Storage Solution 910001
Buy Orion Ph Electrode Storage Solution 910001 Items Including:
Thermo Scientific Orion pH Electrode Storage Solution, 1 pint (475-mL)
Thermo Scientific Orion pH Electrode Storage Solution, 5 x 60-mL bottl
Oakton electrode storage solution, case of 24
Thermo Scientific Orion ROSS Storage Soluton, 475-mL bottle.
More Details
Orion Ph Electrode Storage Solution Msds
Buy Orion Ph Electrode Storage Solution Msds Items Including:
Thermo Scientific Orion pH Electrode Storage Solution, 1 pint (475-mL)
Thermo Scientific Orion pH Electrode Storage Solution, 5 x 60-mL bottl
Oakton electrode storage solution, case of 24
Thermo Scientific Orion ROSS Storage Soluton, 475-mL bottle.
More Details
Cole-Parmer General Purpose Spray Nozzles
Buy Cole-Parmer General Purpose Spray Nozzles Items Including:
PVDF 90° full-cone spray nozzle, 0.053" orifice
PVDF 90° full-cone spray nozzle, 0.074" orifice.
More Details
A&D Weighing Orion Analytical Balances
Buy A&D Weighing Orion Analytical Balances Items Including:
A&D Orion Analytical Balance, 60 g
A&D Orion Analytical Balance, 120 g
A&D Orion Analytical Balance, 210 g
A&D Orion Analytical Balance, 310 g x 0.1 mg, 120 VAC.
More Details
Ohaus Balances
Buy Ohaus Balances Items Including:
Ohaus Scoop, PP
Ohaus Scoop and Counterweight for 01042-01
Ohaus® Compact Scale Adapter; 120 VAC, 50/60 Hz (Accessory)
Ohaus® Compact Scale Container, 1050 mL (Accessory)
Ohaus RS-232 Cable Adapter For Compact Thermal Printer 11901-47 (80251930).
More Details
316l Stainless Steel End Caps
Buy 316l Stainless Steel End Caps Items Including:
End cap, 3/4" (YO-30508-11)
End Cap, 1 - 1 1/2" (YO-30508-12)
316L Stainless Steel Asme-bpe Sanitary Clamp End Cap, 15ra Electropoli (YO-30607-29)
316L Stainless Steel Asme-bpe Sanitary Clamp End Cap, 15ra Electropoli (YO-30607-30).
More Details
Kimble-Chase Long-Tip Measuring (Mohr) Pipettes
Buy Kimble-Chase Long-Tip Measuring (Mohr) Pipettes Items Including:
Kimble-Chase Long-Tip Measuring (Mohr) Pipettes, 1 mL
Kimble-Chase Long-Tip Measuring (Mohr) Pipettes, 2 mL
Kimble-Chase Long-Tip Measuring (Mohr) Pipettes, 5 mL.
More Details
Bronze Gate Valves with Npt(F) Connections
Buy Bronze Gate Valves with Npt(F) Connections Items Including:
Bronze gate valve; 1" NPT(F) (GV0700150)
Bronze gate valve; 1-1/4" NPT(F) (GV0700160).
More Details
Mesh Screen Sieve Set
Buy Mesh Screen Sieve Set Items Including:
Mesh Screen, SS, 1.70 mm sieve opening, 10 mesh
Mesh Screen, SS, 1.40 mm sieve opening, 12 mesh
Mesh Screen, SS, 1.18 mm sieve opening, 14 mesh
Mesh Screen, SS, 1.00 mm sieve opening, 16 mesh
Mesh Screen, SS, 0.850 mm sieve opening, 20 mesh.
More Details
1000 us Cm Conductivity Calibration Solutions
Buy 1000 us Cm Conductivity Calibration Solutions Items Including:
Conductivity calibration standard, 1000 µmhos (3161)
MYRON L conductivity calibration standard, 23.8 µS
MYRON L conductivity calibration standard, 46.7 µS
MYRON L conductivity calibration standard, 2060 µS.
More Details
Radians Safety Glasses
Buy Radians Safety Glasses Items Including:
Radians Journey™ Safety Glasses, Black frame, Clear lens
DeWalt Protector™ Safety Glasses, Clear lens
DeWalt Protector™ Safety Glasses, Smoke lens
DeWalt Contractor Pro™ Safety Glasses, Clear lens
DeWalt Contractor Pro™ Safety Glasses, Smoke lens.
More Details
Electrode for Corning Ph Meter
Buy Electrode for Corning Ph Meter Items Including:
Replacement three-in-one pH electrode, gel-filled junction; 6-ft cable
Replaceable electrode; glass body, refillable, no ATC
pH electrode refillable single-junction for Corning meters (YO-27507-28)
ATC Probe (477968)
Single-junction (YO-27507-20).
More Details
Stainless Steel Tubing Fitting Austin Tx
Buy Stainless Steel Tubing Fitting Austin Tx Items Including:
Compression To Thread Adapter, Stainless Steel, 6mm OD Tubing X 1/4" N (YO-31528-24)
Compression Straight Union, Stainless Steel, 10mm OD Tubing (YO-31528-35)
Compression Reducing Union, Stainless Steel, 12mm X 6mm OD Tubing (YO-31528-41).
More Details
Steel 3 8 Barbed Fitting
Buy Steel 3 8 Barbed Fitting Items Including:
Elbow, Stainless Steel, 3/8", Each (YO-30620-17)
Barbed Fittings; Reducing T Connector, 316 SS, 3/8" x 5/16" Tubing ID (YO-30620-33)
Barbed Fittings; Straight Connector, 316 SS, 3/8" Tubing ID, 1 1/8" Le (YO-31208-06).
More Details
Byko-Charts for Penetration and Opacity
Buy Byko-Charts for Penetration and Opacity Items Including:
byko-charts for Opacity, 5.50" x 10.00", 2A, 250 per box.
More Details
Air Cadet Pumps - Vacuum Pumps
Buy Air Cadet Pumps - Vacuum Pumps Items Including:
Air Cadet Vacuum/Pressure Pump, OEM, 0.65 CFM, 20" HG, 115 VAC
Air Cadet Vacuum/Pressure Pump, Diaphragm, single head, 0.5 cfm, 230 V
Air Cadet® diaphragm vacuum/pressure pump, single head, 0.8 cfm, 1 (YO-07530-25).
More Details
Cole-Parmer Mortar and Pestle Set, Porcelain
Buy Cole-Parmer Mortar and Pestle Set, Porcelain Items Including:
Cole-Parmer Mortar and Pestle Set, Porcelain; 80ML, Each
Cole-Parmer Mortar and Pestle Set, Porcelain; 125ML, Each
Cole-Parmer Mortar and Pestle Set, Porcelain; 300ML, Each.
More Details
Rubber Stopper Size Chart
Buy Rubber Stopper Size Chart Items Including:
Black rubber stopper, size 000
Black rubber stopper, size 00
Black rubber stopper, size 0
Black rubber stopper, size 1
Black rubber stopper, size 2
More Details
70618
results
<<<Previous
25
26
27
28
29
30
31
32
33
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
41
42
43
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Oakton Ph Buffer Coa to Rubber Stopper Size Chart
>
31125329-s-toc_2 31125329stoc2