Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 3
Metrepak Ph
to
Rotary Vane Vacuum Pumps
Image
Product Detail
More Detail
Metrepak Ph/Ion Buffers, and Storage Solutions
Buy Metrepak Ph/Ion Buffers, and Storage Solutions Items Including:
Buffer capsules, pH 4.0, orange (272,PH4)
Buffer capsules, pH 7.0, green (273,PH7)
Buffer capsules, pH 9.0, yellow (270-9.00).
More Details
Tygon Tubing
Buy Tygon Tubing Items Including:
Masterflex® Tygon® tubing, nonphthlate/non-DEHP, 1.85mm ID, 1
Tygon® high-purity tubing, 1/16"ID x 3/16"OD, 50 ft/pack (YO-95635-00)
TYGON Lab Tubing, 4.0 mm ID, 16 ft/pack
Tygon High Purity Tubing, 1/16"ID x 3/16"OD, 50 ft/pk.
More Details
Check Valves with Pipe Thread Connections
Buy Check Valves with Pipe Thread Connections Items Including:
PVC Check Valve, 1/4" NPT(F) connections (CKD025V-PV)
PVC Check Valve, 1/2" NPT(F) connections (CKM050V-PV)
PP Check Valve, 1/4" NPT(F) connections (CKD025V-PP)
PTFE Check Valve, 1/4" NPT(F) connections (CKD025V-TF).
More Details
Ultrasonic Cleaning Solutions
Buy Ultrasonic Cleaning Solutions Items Including:
General-purpose ultrasonic cleaning solution, 1 gallon bottle, pack of (YO-08801-16)
Industrial strength ultrasonic cleaning solution, 5 gallon bottle (YO-08801-27)
Buffing compound remover for ultrasonic cleaners, 1 gallon bottle (000-955-316).
More Details
Compression Fittings
Buy Compression Fittings Items Including:
Inserts for soft tubing for compression fittings, white PP, 5/16" OD x (YO-06391-20)
Compression fittings, male pipe adapter, straight, Black PP, 1/4" OD x (YO-06444-01).
More Details
Cole-Parmer Hazardous-Duty Gear-Drive Air Motor Mixers
Buy Cole-Parmer Hazardous-Duty Gear-Drive Air Motor Mixers Items Including:
Low-cost, gear-drive air motor mixer; 8-170 rpm (YO-04681-00)
Low-cost, gear-drive air motor mixer with steel gears; 50-1200 rpm (YO-04681-10)
Low-cost, gear-drive air motor mixer; 50-1200 rpm (YO-04685-00).
More Details
Stir Bar Retrievers
Buy Stir Bar Retrievers Items Including:
Stir Bar Retriever, Flexible, 14-3/4"L (37774-0101).
More Details
Polypropylene Sanitary Fittings
Buy Polypropylene Sanitary Fittings Items Including:
Polypropylene Sanitary Barbed Adapter, 1" Ladish Clamp Size x 1/4" Tub (YO-31805-31)
Polypropylene Sanitary Barbed Adapter, 1" Ladish Clamp Size x 3/8" Tub (YO-31805-32).
More Details
Globe Valve
Buy Globe Valve Items Including:
Bronze globe valve; 1" NPT(F) (GLV0700750)
Bronze globe valve; 1-1/4" NPT(F) (GLV0700760)
Bronze globe valve; 1-1/2" NPT(F) (GLV0700770).
More Details
Uv Ring Light
Buy Uv Ring Light Items Including:
Black (UV) light ring lights, for 48402-series stereomicroscopes; 115V (YO-41210-00)
Black (UV) light ring lights, for 48403-series stereomicroscopes; 115V (YO-41210-10).
More Details
Stainless Steel Luer Fittings
Buy Stainless Steel Luer Fittings Items Including:
Luer fittings, 316 SS fittings, male luer lock x 1/16" hose barb (YO-31507-26)
Luer fittings, 316 SS fittings, female luer lock x 1/16" hose barb (YO-31507-29)
Stainless steel stopcocksFemale luer x female luer x male luer loc (V5880003).
More Details
Glass Mortar and Pestle
Buy Glass Mortar and Pestle Items Including:
Glass Mortar & Pestle Set, 50ml (YO-34500-51)
Glass Mortar & Pestle Set, 100ml (YO-34500-53)
Glass Mortar & Pestle Set, 250ml (YO-34500-55)
Glass Mortar & Pestle Set, 100ml 6/pk (YO-34501-53).
More Details
Hayward True Union Ball Check Valves
Buy Hayward True Union Ball Check Valves Items Including:
Hayward True Union Ball Check Valve, 1/2" socket and NPT(F) connectors
Hayward True Union Ball Check Valve, 3/4" socket and NPT(F) connectors
Hayward True Union Ball Check Valve, 1-1/2" socket and NPT(F) connecto.
More Details
Asme-Bpe Eccentric Reducer Clamp Fittings
Buy Asme-Bpe Eccentric Reducer Clamp Fittings Items Including:
316L Stainless Steel Asme-bpe Sanitary Clamp Eccentric Reducer, 20ra M (YO-30606-49)
316L Stainless Steel Asme-bpe Sanitary Clamp Eccentric Reducer, 20ra M (YO-30606-50).
More Details
Erlenmeyer Flasks
Buy Erlenmeyer Flasks Items Including:
Erlenmeyer Flask, 10ml/12pk
Erlenmeyer Flask, 25ml/12pk
Kimax Erlenmeyer Flask with Screw Caps, 250mL (YO-34524-41)
Kimax Erlenmeyer Flask with Screw Caps, 500mL (YO-34524-43)
Erlenmeyer flask mantle for 125-ml flask, 95 watts, 230 VAC (100AO913).
More Details
Parker Hannifin Bulkhead Union Fittings
Buy Parker Hannifin Bulkhead Union Fittings Items Including:
Compression fittings, Bulkhead unions; Black PP, 3/8" OD, 2-5/8" x 15/ (YO-06444-32)
Compression bulkhead union, PFA, 1/8 (YO-31320-82)
Compression bulkhead union, PFA, 1/4 (YO-31320-83)
Compression bulkhead union, PFA, 3/8 (YO-31320-84).
More Details
Kimax Graduated Cylinders
Buy Kimax Graduated Cylinders Items Including:
100 mL Kimax class B graduated cylinders
250 mL Kimax class B graduated cylinders
500 mL Kimax class B graduated cylinders
1000 mL Kimax class B graduated cylinders
2000 mL Kimax class B graduated cylinders
More Details
Parker Three-Way Ball Valves
Buy Parker Three-Way Ball Valves Items Including:
Parker three-way ball valve, stainless steel, 1/8" NPT(F)
Parker three-way ball valve, stainless steel, 1/4" NPT(F)
Parker three-way ball valve, brass, 1/8" compression connection
Parker three-way ball valve, brass, 1/8" NPT(F).
More Details
Specwell Corporation Microscope/Telescopes with Built-in Reticles
Buy Specwell Corporation Microscope/Telescopes with Built-in Reticles Items Including:
Microscope/telescopes with built-in reticle; magnification, 25x/8x; fi (M820-S)
Microscope/telescopes with built-in reticle; magnification, 25x/8x; fi (M830-S)
Microscope/telescopes with built-in reticle; magnification, 25x/8x; fi (M820-SM).
More Details
Steel Tubing
Buy Steel Tubing Items Including:
Steel tubing clamp for 3/4"OD tubing (YO-06404-30)
316 Stainless Steel Tubing, high pressure; 1/16" OD; two 3-ft sections (YO-03300-02)
316 Stainless Steel Tubing; dimensions, 1/16" ID, 1/8" OD, 1/32" wall; (YO-03300-05).
More Details
Mark-10 Inline Tension/Compression Force Sensors
Buy Mark-10 Inline Tension/Compression Force Sensors Items Including:
Mark-10 MR03-025 Force Sensor, .25 lb x 100 g x 1 N
Mark-10 MR03-20 Force Sensor, 20 lb x 10 kg x 100 N
Mark-10 MR03-50 Force Sensor, 50 lb x 25 kg x 250 N
Mark-10 MR03-05 Force Sensor, .5 lb x 250 g x 2.5 N.
More Details
Mark-10 S-Beam Tension/Compression Force Sensors
Buy Mark-10 S-Beam Tension/Compression Force Sensors Items Including:
Mark-10 MR01-200 Force Sensor, 200 lb x 100 kg x 1000 N
Mark-10 MR01-1000 Force Sensor, 1000 lb x 500 kg x 5000 N
Mark-10 MR01-2000 Force Sensor, 2000 lb x 1000 kg x 10000 N
Mark-10 MR01-10000 Force Sensor, 10000 lb x 5000 kg x 50 kN.
More Details
Industrial Strength Cleaning Solution
Buy Industrial Strength Cleaning Solution Items Including:
Industrial strength ultrasonic cleaning solution, 5 gallon bottle (YO-08801-27).
More Details
Glass Bottles
Buy Glass Bottles Items Including:
Cole-Parmer Bottletop Dispenser .25-2.5 mL with 100 mL Glass Bottle
Boston Round Glass Bottle Mailer, 1 oz, 3-3/4" L x 1-1/2" dia, 24/cs (2300)
Medium Round Glass Bottle Mailer, 1 oz, 3-3/4" L x 1-1/2" dia, 24/cs (2304).
More Details
Whatman Filter Paper Pore Size 2
Buy Whatman Filter Paper Pore Size 2 Items Including:
Whatman Prepleated Filter Papers; 12.5 cm dia; pore size, 8 µ; 100/
Whatman Prepleated Filter Papers; 15 cm dia; pore size, 8 µ; 100/bo
Whatman Prepleated Filter Papers; 18.5 cm dia; pore size, 8 µ; 100/.
More Details
Hazardous-Duty Gear-Drive Air Motor Mixers
Buy Hazardous-Duty Gear-Drive Air Motor Mixers Items Including:
Low-cost, gear-drive air motor mixer; 8-170 rpm (YO-04681-00)
Low-cost, gear-drive air motor mixer with steel gears; 50-1200 rpm (YO-04681-10)
Low-cost, gear-drive air motor mixer; 50-1200 rpm (YO-04685-00).
More Details
Lmi Metering Pumps
Buy Lmi Metering Pumps Items Including:
LMI Solenoid Metering Pump Service Kit for 74522-*6
LMI Solenoid Metering Pump Service Kit for 74522-*8
Digital Remote-Control Solenoid-Diaphragm Metering Pump, 2.5 GPH, 115
Digital Remote-Control Solenoid-Diaphragm Metering Pump, 0.42 GPH, 115.
More Details
Thermo Scientific Nalgene Media Bottles
Buy Thermo Scientific Nalgene Media Bottles Items Including:
Thermo Scientific Nalgene square polycarbonate bottle, 125 mL
Thermo Scientific Nalgene square polycarbonate bottle, 1 L
Thermo Scientific Nalgene square polycarbonate bottle, 2 L
Thermo Scientific Nalgene Sterile PETG Media Bottles, 30 mL; 96/box.
More Details
Interscience Bagmixer Paddle Blender Bags
Buy Interscience Bagmixer Paddle Blender Bags Items Including:
Bagpage Paddle Blender Bags, 80ml With Full Filter, Sterile Packs Of 2 (120-825)
Bagpage Flat Bottom Paddle Blender Bags, 400ml With Full Filter, 280 m (122-025)
Bagpage Flat Bottom Paddle Blender Bags, 400ml With Full Filter, 63 mi (122-325).
More Details
Rotary Vane Vacuum Pumps
Buy Rotary Vane Vacuum Pumps Items Including:
Vacuubrandreg Rotary Vane Vacuum Pump, 6.0 cfm / 165 lpm, 1.5 X 10s
Cole-Parmer® Two Stage Rotary Vane Vacuum Pump, 106/128L/min, 3x10 (YO-72226-65)
Dry rotary vane vacuum pump, 4.5 cfm, 115 VAC (0523-V542AQ-G588DX)
Vacuum Pump 96 liters/min, 230V.
More Details
70618
results
<<<Previous
1
2
3
4
5
6
7
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Metrepak Ph to Rotary Vane Vacuum Pumps
>
31125329-s-toc_2 31125329stoc2