Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 300
Protimeter General Accessories
to
Honeywell Ph Meter More Categories
Image
Product Detail
More Detail
Protimeter General Accessories
Buy Protimeter General Accessories Items Including:
BLD5070 : EIFS Probe
Gateway
BLD9050 : Wireless Sensor
BLD4750C : Hygrosticks, 5 Pack
50-Pack
More Details
Small Glass Vials with Screw Top
Buy Small Glass Vials with Screw Top Items Including:
LVI 0.3 mL Clear Vial with 9mm Blue Screw Cap, PTFE/Sil, 100/CS
LVI 0.3 mL Amber Vial with 9mm Blue Screw Cap, PTFE/Sil, 100/CS
Closures for Screw-Thread Vials; Top seal, clear molded 10 mil PP, 8-4 (YO-98721-10).
More Details
Eutech Instruments Ph/Ion Handheld Meters
Buy Eutech Instruments Ph/Ion Handheld Meters Items Including:
Eutech pH 6+ pH/ORP meter kit (YO-15940-80).
More Details
Vacuum Pump 30 Hg
Buy Vacuum Pump 30 Hg Items Including:
Venturi vacuum pump, 1.2 cfm, 29.5
PTFE Vacuum Pump, 70 L/min free air capacity, 28.7" Hg (30 torr) 115 V
PTFE Vacuum Pump, 70 L/min free air capacity, 28.7" Hg (30 torr) 230 V
Cole-Parmer Vacuum Pump, 0.31 cfm, 21.3" Hg, 230 VAC.
More Details
Nichiryo Pipettes and Pipettors
Buy Nichiryo Pipettes and Pipettors Items Including:
Nichiryo Le Adjustable-Volume Pipettor, 100 to 1000 µL (00-NLE-1000)
Repetitive syringe dispenser (NSTP)
Syringe, 0.05 mL tip volume (NS-V)
Syringe, 0.5 mL tip volume (NS-S)
Syringe, 1.25 mL tip volume (NS-M)
More Details
Applikon Deep-Well Microflask Systems
Buy Applikon Deep-Well Microflask Systems Items Including:
Applikon Deep-Well MicroFlask Systems Microtiter 24-well deep-well pol
Applikon Deep-Well MicroFlask Systems Microtiter 96-well deep-well pol
Applikon Pre-Sterilized Micro-Titer Plate for 24 Deep-Well Square, pyr
24 Deep-Well silicone gasket/seal.
More Details
3m Bx Dual Reader Safety Glasses
Buy 3m Bx Dual Reader Safety Glasses Items Including:
3M BX™ Dual Reader Safety Glasses, 2.0X top and bottom diopters
3M BX™ Dual Reader Safety Glasses, 2.5x top and bottom diopters.
More Details
Nonsterile Culture Tubes without Caps
Buy Nonsterile Culture Tubes without Caps Items Including:
Nonsterile culture tubes without cap; 17mm x 100mm; material, PP; colo
Nonsterile culture tubes without cap; 17mm x 100mm; material, PS; colo
Nonsterile culture tubes without caps; material, color, clear; PS; 12 (8560-1).
More Details
Respirator P100
Buy Respirator P100 Items Including:
Sperian Saf-T-Fit Plus Particulate Respirators; P100, Exhalation Valve (YO-81660-27)
Filter cartridge with P100 filter; UNISORB
Filter cartridge, Multigas and vapor; P100 (YO-40113-75)
Filter cartridge with P100 filter; Organic vapor (YO-81661-67).
More Details
Centrifuge Tube Insert
Buy Centrifuge Tube Insert Items Including:
Centrifuge Tube Insert for Automatic Heated Oil Test Rotor, 100 mL Lon (YO-17300-25)
Centrifuge Tube Insert for Automatic Heated Oil Test Rotor, 100 mL Lon (YO-17300-27)
Centrifuge Tube Insert for Automatic Heated Oil Test Rotor, 100 mL Con.
More Details
Ika Microplate Shakers
Buy Ika Microplate Shakers Items Including:
Digital Shaker 115v (MTS 2/4)
Microplate shaker
Separating Funnel Attachment for IKA Digital Shakers, for 12 x 50-mL,
Separating Funnel Attachment for IKA Digital Shakers, for 4 x 500-mL,
MicroPlate Genie Shaker 120V
More Details
Colder Hose Barb Elbow Inserts
Buy Colder Hose Barb Elbow Inserts Items Including:
Quick-Disconnect Hose Barb Elbow Insert; Straight-through, Delrin, 1/8 (PMC2302)
Quick-Disconnect Hose Barb Elbow Insert; Straight-through, Delrin, 1/4 (PMC2304)
Quick-Disconnect Hose Barb Elbow Insert; Valved, Delrin, 1/4" Tube ID, (PMCD2304).
More Details
Hdpe Tanks
Buy Hdpe Tanks Items Including:
Cylidrical Tank with Cover, HDPE, 100 Gal
(YO-06317-10)
Cylidrical Tank with Cover, HDPE, 150 Gal
(YO-06317-20)
Cylidrical Tank with Cover, HDPE, 200 Gal
More Details
Click N Go General Accessories
Buy Click N Go General Accessories Items Including:
Click'n'go™ replacement three-stop minicartridge.
More Details
Cole-Parmer Portable Benchtop Chilling Incubator
Buy Cole-Parmer Portable Benchtop Chilling Incubator Items Including:
Portable Benchtop Chilling Incubator, 100/240VAC (YO-39064-21).
More Details
Humidity Calculator
Buy Humidity Calculator Items Including:
Humidity Slide Calculator (HC880).
More Details
Lab Products Hybridization Incubators
Buy Lab Products Hybridization Incubators Items Including:
Glass Hybridization Bottle, Large, 25x300mm (YO-39453-22).
More Details
Liquid Dispensers in Laboratories
Buy Liquid Dispensers in Laboratories Items Including:
Polyethylene liquid dispenser, 3 gallon (H0121203)
Polyethylene liquid dispenser, 6 gallon (H0240603).
More Details
Nalgene Carboy 2302 0005
Buy Nalgene Carboy 2302 0005 Items Including:
Thermo Scientific Nalgene low-density polyethylene carboy with tubulat (2302-0020).
More Details
Qrp Latex Antistatic Finger Cots
Buy Qrp Latex Antistatic Finger Cots Items Including:
QRP™ Latex Antistatic Finger Cots, small
QRP™ Latex Antistatic Finger Cots, medium
QRP™ Latex Antistatic Finger Cots, large.
More Details
Turbine Propellers, 304 Stainless Steel
Buy Turbine Propellers, 304 Stainless Steel Items Including:
304 SS turbine propeller for shaft 04370-02, 100 mm propeller dia (YO-04370-16).
More Details
1000 Psi Pressure Gauge
Buy 1000 Psi Pressure Gauge Items Including:
0 to 1000 psi Industrial Gauge with Bottom Connection, 2 1/2" Dial
0 to 1000 psi Industrial Gauge with Back Connection, 2 1/2" Dial
Ashcroft Pressure Gauge Comparator, 1000 psi
Dwyer Magnehelic Differential Pressure Gauge, 2210: 0/10 psi.
More Details
Horiba Conductivity Meter B 173 Manu
Buy Horiba Conductivity Meter B 173 Manu Items Including:
Replacement conductivity cell for Horiba conductivity and multiparamet
Horiba DS-72 conductivity benchtop meter
Horiba F-74 pH/Conductivity benchtop meter
Horiba F-72 pH/ORP/ION benchtop meter
Horiba B-171 Twin Conductivity/Salinity Pocket Tester.
More Details
Gilson Ss 15 Sieve Shaker
Buy Gilson Ss 15 Sieve Shaker Items Including:
Vibro-Separator, Nonportable, SS Model, 230 VAC
Mesh Screen, SS, 1.70 mm sieve opening, 10 mesh
Mesh Screen, SS, 1.40 mm sieve opening, 12 mesh
Mesh Screen, SS, 1.18 mm sieve opening, 14 mesh
Mesh Screen, SS, 1.00 mm sieve opening, 16 mesh.
More Details
Spectra/Por Standard Rc Dialysis Trial Kits
Buy Spectra/Por Standard Rc Dialysis Trial Kits Items Including:
Spectra/Por Dialysis Membrane Trial Kit, 6000-8000Dalton 32mm
Spectra/Por Dialysis Membrane Trial Kit, 10,000-12,000Dalton 25mm
Spectra/Por Dialysis Membrane Trial Kit, 10,000-12,000Dalton 50mm.
More Details
Accumet Conductivity Meter
Buy Accumet Conductivity Meter Items Including:
accumet® AB 30 benchtop conductivity meter kit
accumet® XL 30 benchtop conductivity meter only
Accumet XL200 pH/Conductivity Benchtop Meter Only
Accumet XL200 pH/Conductivity Benchtop Meter Kit
Accumet XL500 pH/ISE/Conductivity Benchtop Meter Kit.
More Details
Eccentric Tubing Reducers
Buy Eccentric Tubing Reducers Items Including:
ECCENTRIC SANITARY CLAMP REDUCER, 316L SS, 1-1/2" X 1" (YO-30560-05)
ECCENTRIC SANITARY CLAMP REDUCER, 316L SS, 2" X 1-1/2" (YO-30560-07)
ECCENTRIC SANITARY CLAMP REDUCER, 304L SS, 1-1/2" X 1" (YO-30561-05).
More Details
Brass Astm Mortar Testing Sieve
Buy Brass Astm Mortar Testing Sieve Items Including:
Cole-Parmer Testing Sieve, 8" OD Brass Frame/Brass Wire, Full height, (YO-59980-00)
Cole-Parmer Testing Sieve, 8" OD Brass Frame/Brass Wire, Full height, (YO-59980-01).
More Details
Accessories for Flowmeters and Sensors
Buy Accessories for Flowmeters and Sensors Items Including:
Replacement 4.5 mm PVDF Turbine Flow Sensor for PP Tube Holder 32516-3
Sensor Cap for In-line Mounting, Red (YO-05614-27)
Cable, 10-ft length with 3-pin connector
Connects flow sensors 32249- (HB220-219)
Standard sensor – 2 wire sine wave output.
More Details
Honeywell Ph Meter More Categories
Buy Honeywell Ph Meter More Categories Items Including:
Honeywell Directline ® pH Transmitter Electronics Module
Honeywell Durafet III, non-glass pH electrode, 3/4 inch immersion thre
Honeywell Durafet III, non-glass pH electrode, 3/4 inch in-line thread
Honeywell dissolved oxygen input card, ppm range.
More Details
70618
results
<<<Previous
296
297
298
299
300
301
302
303
304
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
100
200
210
220
230
240
250
260
270
280
285
286
287
288
289
290
291
292
293
294
295
296
297
298
299
300
301
302
303
304
305
306
307
308
309
310
311
312
313
314
320
330
340
350
360
370
380
390
400
500
600
700
800
900
1,000
1,100
1,200
2,000
2,354
Protimeter General Accessories to Honeywell Ph Meter More Categories
>
31125329-s-toc_2 31125329stoc2