Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 4

Brass Check Valves   to   Thermo Scientific Revco Laboratory Refrigerators

Image
  Product Detail
More Detail

Brass Check Valves

Buy Brass Check Valves Items Including:
  • Brass Check Valve, 1/4" NPT(F) connectors (0376369)
  • Brass Check Valve, 3/8" NPT(F) connectors (0376370)
  • Brass Check Valve, 1/2" NPT(F) connectors (0376371)
  • Brass Check Valve, 3/4" NPT(F) connectors (0376372).

Sartorius Balance and Scale Accessories

Buy Sartorius Balance and Scale Accessories Items Including:
  • RS-232 Adapter for Sartorius Element balances
  • USB Adapter for Sartorius Element balances
  • 220 VAC Power adapter with out cord for Sartorius LE, CP, ED, EA/EB an
  • RS-232 Interface (YDO01BL)
  • Printer,

Tube Racks

Buy Tube Racks Items Including:
  • 10-Channel Tube Rack, 3/16, 10/pk (YO-06432-02)
  • 10-Channel Tube Rack, 1/4, 10/pk (YO-06432-03)
  • Test tube racks for 1.5 mL tube size; PP (YO-06703-40)
  • Test tube racks for 0.5 mL tube size; PP (YO-06703-41)
  • Poxygrid Wire Test Tube Rack, 60, 18-20mm.

Abb Recorder Chart Paper

Buy Abb Recorder Chart Paper Items Including:
  • Chart Paper for ABB Commander 250-mm Strip Chart Recorders, 80 divisio
  • Chart Paper for ABB Commander Circular Chart Recorder/Controllers, 0 t (500P1225-106)
  • Chart Paper for ABB Commander Circular Chart Recorder/Controllers, 0 t (500P1225-15).

Troemner Gas Cylinder Securing Safety Straps

Buy Troemner Gas Cylinder Securing Safety Straps Items Including:
  • Bench Clamp with secure message (973036)
  • Wall Bracket with secure message (973037)
  • Bench Clamp with secure message (974036)
  • Wall Bracket with secure message (974037)
  • Safety Strap without Message

Kimax Glass Volumetric Flasks

Buy Kimax Glass Volumetric Flasks Items Including:
  • KIMAX Square Volumetric Flask, 100mL 1/pk
  • KIMAX Square Volumetric Flask, 200mL 1/pk
  • KIMAX Square Volumetric Flask, 250mL 1/pk
  • Kimax Wide-Mouth Volumetric Flask, 5mL, Glass stopper, 6/cs
  • Kimax Wide-Mouth Volumetric Flask, 10mL, Glass stopper, 6/cs.

Ashcroft Pressure Gauge

Buy Ashcroft Pressure Gauge Items Including:
  • Ashcroft Pressure Gauge Comparator, 0-100 psi
  • Ashcroft Pressure Gauge Comparator, 1000 psi
  • Ashcroft Pressure Gauge Comparator, 5000 psi
  • Ashcroft Pressure Gauge Comparator, 10,000 psi
  • Ashcroft Dual-Scale Low-Pressure Gauge, 0 to 10" H2O and 0.

A&D Weighing Orion Hr Analytical Balances

Buy A&D Weighing Orion Hr Analytical Balances Items Including:
  • A&D Orion Analytical Balance, 60 g
  • A&D Orion Analytical Balance, 120 g
  • A&D Orion Analytical Balance, 210 g
  • A&D Orion Analytical Balance, 310 g x 0.1 mg, 120 VAC.

Norprene Tubing

Buy Norprene Tubing Items Including:
  • Norprene Tubing, 1/16"ID x 3/16"OD, 50 ft/pack
  • Norprene Tubing, 3/16"ID x 5/16"OD, 50 ft/pack
  • Norprene Tubing, 1/4"ID x 1/2"OD, 50' per pack
  • Norprene Tubing, 5/16"ID x 7/16"OD, 50 ft/pack.

Amber Glass Bottles

Buy Amber Glass Bottles Items Including:
  • Bottle, precleaned, amber glass, narrow mouth, Qorpak, 60 mL, 20 mm ne
  • Bottle, precleaned, amber glass, narrow mouth, Qorpak, 120 mL, 22 mm n
  • 30 mL Amber Glass Dropping Bottle, Glass dropper
  • Amber glass dropping bottle, 50 mL
  • Pack of 6

Clear Plastic Storage Bins

Buy Clear Plastic Storage Bins Items Including:
  • Storage bin; 4-1/8" x 3" x 5-3/8", 24/pack (30-210 YELLOW)
  • Storage bin; 4-1/8" x 3" x 7-3/8", 24/pack (30-220 YELLOW)
  • Storage bin; 5-1/2" x 5" x 10-7/8", 12/pack (30-230 YELLOW).

Thermo Scientific Barnstead Nanopure Cartridge Kits

Buy Thermo Scientific Barnstead Nanopure Cartridge Kits Items Including:
  • Organic-Free RO or Distilled Feed Cartridge Pack for Thermo Scientific (D50280)
  • Thermo Scientific Barnstead Type I cartridge kit, organic free, four m (D5023)
  • Thermo Scientific Barnstead Type I Cartridge Kit, for tap or deionized (D50256).

Specific Gravity Cups

Buy Specific Gravity Cups Items Including:
  • Specific Gravity Cup, 83.2 mL capacity, 0.5% volume tolerance, for use (PV-9654)
  • Specific gravity cup, 8.32 mL capacity, 1.2% volume tolerance, for use (WG-SS-8.32)
  • Specific gravity cup, 83.2 mL capacity, 0.5% volume tolerance, for use (WG-SS-83.2).

Hot Plates

Buy Hot Plates Items Including:
  • Cole-Parmer Five-Position Stirring Hot Plate, 120 VAC
  • Cole-Parmer Nine-Position Stirring Hot Plate, 120 VAC
  • Cole-Parmer Five-Position Stirring Hot Plate, 230 VAC
  • EchoTherm™ Digital Stirring Hot Plate , 115 VAC
  • Basic Mini Stirring Hot Plate, 120V.

Wheaton Glass Bottles

Buy Wheaton Glass Bottles Items Including:
  • WHEATON BOD Bottle, 300 mL, glass, unnumbered, penny head stopper, 24/
  • WHEATON BOD Bottle, 300 mL, glass, unnumbered, robotic stopper, 24/cas
  • Clear glass dropping bottle, 100 mL
  • Pack of 6
  • (W211735)

Glassine Weighing Paper

Buy Glassine Weighing Paper Items Including:
  • Glassine Weighing Paper 3x3" 500/pk (YO-01338-00)
  • Glassine Weighing Paper 4x4" 500/pk (YO-01338-02).

Plastic Volumetric Flask

Buy Plastic Volumetric Flask Items Including:
  • Volumetric Flask, PMP, 25mL, 1/pk (YO-06110-32)
  • Volumetric Flask, PMP, 50mL, 1/pk (YO-06110-34)
  • Volumetric Flask, PMP, 100mL, 1/pk (YO-06110-36)
  • Volumetric Flask, PMP, 250mL, 1/pk (YO-06110-38)
  • Volumetric Flask, PMP, 500mL, 1/pk (YO-06110-40).

Nalgene Laboratory Wash Bottles

Buy Nalgene Laboratory Wash Bottles Items Including:
  • Thermo Scientific Nalgene PPCO wash bottle, 500 mL
  • Thermo Scientific Nalgene PPCO wash bottle, 1 L
  • Thermo Scientific Nalgene wide-mouth Unitary wash bottle, 125 mL
  • Thermo Scientific Nalgene "Right-to-Know" safety wash bottle for aceto.

Stainless Steel Funnels

Buy Stainless Steel Funnels Items Including:
  • Stainless steel funnel, 2 3/8 oz (YO-07263-00)
  • Stainless steel funnel, 13 oz (YO-07265-00)
  • Funnel rack for modular stainless steel drying racks
  • Justrite Safety Drum Funnel, manual closing, 1" stainless steel flame
  • 150ml Funnel, Kg25 (311208).

Phydrion Ph Test Strips and Indicators

Buy Phydrion Ph Test Strips and Indicators Items Including:
  • pHydrion Insta-Chek Mechanical pH Pencil, pH 0-13, 3 per package
  • pHydrion plastic pH indicator strips, 0.0 to 14.0, flip top vial packa
  • pHydrion plastic pH indicator strips, 0.0 to 6.0, flip top vial packag
  • Hydrion pH Spill Stick, 0-13 pH, 10/pk.

Nikon General Microscope Accessories

Buy Nikon General Microscope Accessories Items Including:
  • 10:1 Stereozoom Microscope 35-mm Camera Adapter, Nikon (T2-5).

3m Respirator Cartridge

Buy 3m Respirator Cartridge Items Including:
  • Filter cartridge, Organic vapor (YO-40113-50)
  • Filter cartridge, Organic vapor/Acid gas (YO-40113-52)
  • Filter cartridge, Multigas and vapor (YO-40113-55)
  • Filter cartridge, Organic vapor/Acid gas; P100 (YO-40113-72)
  • Cartridge; Acid gas (YO-40120-51).

Duct Tape

Buy Duct Tape Items Including:
  • 3M Value Duct Tape 1900 Silver, 1.88" x 50yd (70-0064-1242-6)
  • 3M Duct Tape 1900 Silver, 2.83" x 50yd (70-0064-1243-4)
  • 3M Duct Tape 3939 Silver, 1.88" x 50yd (70-0062-5013-1).

Thermo Scientific Nalgene Hdpe Sample Bottles

Buy Thermo Scientific Nalgene Hdpe Sample Bottles Items Including:
  • Thermo Scientific Nalgene high-density polyethylene narrow-mouth sampl (2089-0001)
  • Thermo Scientific Nalgene high-density polyethylene narrow-mouth sampl (2089-0002)
  • Thermo Scientific Nalgene high-density polyethylene narrow-mouth sampl (2089-0004).

Ph Test Strips for Water

Buy Ph Test Strips for Water Items Including:
  • EM Quant pH test strips, 0-14 pH
  • Six packs of 100.
  • pHydrion plastic pH indicator strips, 0.0 to 14.0, flip top vial packa
  • pHydrion plastic pH indicator strips, 0.0 to 6.0, flip top vial packag
  • EM Quant® Zinc Test Strips (10038-1).

Zirconia Grinding Media

Buy Zirconia Grinding Media Items Including:
  • Zirconia grinding media, 3/8" x 3/8" (YO-04158-10)
  • Zirconia grinding media, 1/2" x 1/2" (YO-04158-20)
  • Grind Media, Zirconia, 2425/pk (YO-04158-00).

Abb Commander Circular Chart Recorder/Controllers

Buy Abb Commander Circular Chart Recorder/Controllers Items Including:
  • ABB Commander Circular Chart Recorder/Controller - 1 controller, 2 pen
  • Chart Paper for ABB Commander Circular Chart Recorder/Controllers, 0 t (500P1225-106)
  • Chart Paper for ABB Commander Circular Chart Recorder/Controllers, 0 t (500P1225-15).

Polypropylene Jars

Buy Polypropylene Jars Items Including:
  • Thermo Scientific Nalgene straight-sided polypropylene wide-mouth jar, (2118-9050)
  • Thermo Scientific Nalgene straight-sided polypropylene wide-mouth jar, (2118-0001)
  • Thermo Scientific Nalgene straight-sided polypropylene wide-mouth jar, (2118-0002).

Hvac/R Replacement Heating Element Kits

Buy Hvac/R Replacement Heating Element Kits Items Including:
  • Replacement heating elements for heat sealer, 8", 115 VAC (K200H)
  • Replacement heating elements for heat sealer, 8", 230 VAC (K205H)
  • Replacement heating elements for heat sealer, 12", 115 VAC (K300H).

Thermo Scientific Revco Laboratory Refrigerators

Buy Thermo Scientific Revco Laboratory Refrigerators Items Including:
  • Laboratory undercounter refrigerator (single door); 5.4 CU
  • FT; 115V (REL-404A)
  • Laboratory space-saver refrigerator (single door); 11.5 CU FT; 115V (REL-1204A-20)
  • Laboratory space-saver refrigerator (single door); 11.5 CU FT; 200/230 (REL-1204D).

70618 results


Brass Check Valves to Thermo Scientific Revco Laboratory Refrigerators

>