Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 400

Love Temperature Controller Vendors   to   Kelway Soil Ph Meter

Image
  Product Detail
More Detail

Love Temperature Controller Vendors

Buy Love Temperature Controller Vendors Items Including:
  • Love Temperature Control, 1/32 DIN, Universal Input, Voltage Pulse & R (32B-23)
  • Love Temperature Control, 1/32 DIN, Universal Input, Relay Outputs, Ra (32B-33)
  • Love Temperature Control, 1/16 DIN, Universal Input, Voltage Pulse & R (16B-23).

Love Temperature Controllers

Buy Love Temperature Controllers Items Including:
  • Love Temperature Control, 1/32 DIN, Universal Input, Voltage Pulse & R (32B-23)
  • Love Temperature Control, 1/32 DIN, Universal Input, Relay Outputs, Ra (32B-33)
  • Love Temperature Control, 1/16 DIN, Universal Input, Voltage Pulse & R (16B-23).

Utility Trays

Buy Utility Trays Items Including:
  • UltraTech Utility Tray, 12" x 48" x 4-3/4", 12 gallon capacity
  • Stainless steel utility tray, 10"L x 6-1/2"W
  • Stainless steel utility tray, 13-1/2"L x 9-3/4"W
  • Stainless steel utility tray, 17"L x 11-5/8"W.

Dew Point Temperature Humidity Calculator

Buy Dew Point Temperature Humidity Calculator Items Including:
  • Rh / Dew Point And Temperature Transmitter (no Display - Remote Probe) (DMT340-8V0B0A1BEA1B001A1ACB0A0)
  • Dew Point Meter (DPMi)
  • GE (General Eastern) Industrial Dew Point Transmitter with Display
  • Industrial dew Point Transmitter with Plastic Housing.

Cole-Parmer Continuous Alarm Timer

Buy Cole-Parmer Continuous Alarm Timer Items Including:
  • Cole-Parmer Continuous Visual Alarm Timer.

Conductivity & Ph Measurement

Buy Conductivity & Ph Measurement Items Including:
  • Oakton® high-accuracy 10.000 pH buffer, 500-mL, case of 12
  • 100 Ω Pt RTD Conductivity Cell for ultra-low range measurements, (YO-19500-02)
  • 100 Ω Pt RTD Conductivity Cell for ultra-low range measurements, (YO-19500-04).

Luer Type 316 Stainless Steel Adapters

Buy Luer Type 316 Stainless Steel Adapters Items Including:
  • 316 Stainless Steel fittings, female luer x female luer (YO-31507-89)
  • Luer fittings, 316 SS fittings, female luer x male slip (YO-31507-20)
  • 316 SS fittings; female luer x male luer lock (YO-31507-21)
  • 316 SS fittings; male slip plug (YO-31507-22).

Limit Alarms and Signal Conditioners

Buy Limit Alarms and Signal Conditioners Items Including:
  • I/q Rail AC Pwr Dist Asy 2mod
  • I/q Rail DC Pwr Dist Asy 4mod
  • I/q Rail AC Pwr Dist Asy 4mod
  • I/q Rail DC Pwr Dist Asy 8mod.

Thermo Scientific Orion Orp Standards

Buy Thermo Scientific Orion Orp Standards Items Including:
  • Thermo Scientific Orion ORP Standard, one 475-mL bottle.

Plastic Water Pump

Buy Plastic Water Pump Items Including:
  • Hand-operated water and chemical siphon/drum pump, 32 strokes/gallon
  • Hand-operated chemical siphon/drum pump, 12 strokes/gallon.

Stainless Steel Ultrasonic Cleaner with Timers

Buy Stainless Steel Ultrasonic Cleaner with Timers Items Including:
  • Cole-Parmer Stainless Steel Ultrasonic Cleaner with Mechanical Timer, (YO-08895-00)
  • Cole-Parmer Stainless Steel Ultrasonic Cleaner with Mechanical Timer, (YO-08895-06).

Cole-Parmer Wilmad-Labglass Tlc Developing Cylinders

Buy Cole-Parmer Wilmad-Labglass Tlc Developing Cylinders Items Including:
  • TLC Complete Developing Tank For 25 X 75mm Plates (YO-34107-00)
  • TLC Complete Developing Tank For 50 X 100mm Plates (YO-34107-20).

Myron L Conductivity Solutions

Buy Myron L Conductivity Solutions Items Including:
  • MYRON L conductivity calibration standard, 23.8 µS
  • MYRON L conductivity calibration standard, 46.7 µS
  • MYRON L conductivity calibration standard, 2060 µS
  • MYRON L conductivity calibration standard, 16,630 µS.

Oakton Ecotestr Ph 2

Buy Oakton Ecotestr Ph 2 Items Including:
  • Oakton Waterproof EcoTestr™ pH 2 Pocket pH Tester
  • Oakton EcoTestr TDS High pocket TDS tester
  • Oakton EcoTestr EC Low pocket conductivity tester
  • Oakton EcoTestr EC High pocket conductivity tester
  • Oakton EcoTestr Salt pocket salinity tester.

Thermo Scientific Sorvall St 40 Centrifuges

Buy Thermo Scientific Sorvall St 40 Centrifuges Items Including:
  • Thermo Scientific Sorvall ST 40 Centrifuge, 120VAC
  • Thermo Scientific Sorvall ST 40 Centrifuge, 230VAC
  • Thermo Scientific Sorvall ST 40R Refrigerated Centrifuge, 120VAC
  • Thermo Scientific Bioliner High Capacity Rotor, 4 Positions; Requires (75003667).

Threaded Pipe Bushing

Buy Threaded Pipe Bushing Items Including:
  • Pipe fittings, Reducing bushing NPT(M) x NPT(F), Brass, 1/4" x 1/8", 7 (YO-30912-56)
  • Pipe fittings, Reducing bushing NPT(M) x NPT(F), Brass, 3/8" x 1/8", 7 (YO-30912-57).

Sintered Funnel

Buy Sintered Funnel Items Including:
  • Scienceware Separatory Funnel Rack
  • Separating Funnel Attachment for IKA Digital Shakers, for 12 x 50-mL,
  • Cole-Parmer Utility Funnel, 5mL, 12/pk
  • Cole-Parmer Utility Funnel, 25mL, 12/pk
  • Cole-Parmer Utility Funnel, 40 mL, 12/pk

Accuform Danger Hard Hat Area Signs

Buy Accuform Danger Hard Hat Area Signs Items Including:
  • Safety Sign, Danger - Hard Hat Area, 7" X 10", Adhesive Vinyl
  • Safety Sign, Danger - Hard Hat Area, 7" X 10", Aluminum
  • Safety Sign, Danger - Hard Hat Area, 10" X 14", Adhesive Vinyl.

Air Compressor Air Tools

Buy Air Compressor Air Tools Items Including:
  • Air Tool, Butterfly air impact wrench, 3/8 in drive, 3/8 in bolt capac (AT-5030)
  • Air Tool, Air impact wrench, 3/4 in drive, 1 in bolt capacity, 5,000 r (AT-5060L)
  • Air Tool, Air impact wrench, 1 in drive, 8 in Anvil, 1-1/2 in bolt cap (AT-5281L).

Compressor and Air Tools

Buy Compressor and Air Tools Items Including:
  • Air Tool, Butterfly air impact wrench, 3/8 in drive, 3/8 in bolt capac (AT-5030)
  • Air Impact Wrench, 1/2" drive, 9800 rpm
  • Air Impact Wrench, 1/2" drive, 8000 rpm
  • Vacuum-Ready Random Orbital Sander Kit, Air powered, 6" pad size, 12,0.

Keyed Mixer Chucks

Buy Keyed Mixer Chucks Items Including:
  • Mixer chuck; keyed (YO-04552-86)
  • Mixer chuck and key set (YO-04552-88)
  • Chuck key for 04552-86 (YO-04552-87).

Thermo Scientific Nalgene Bottles

Buy Thermo Scientific Nalgene Bottles Items Including:
  • Flexible Top Works™ Closures for Thermo Scientific Nalgene Bottles (2135-3803)
  • Flexible Top Works™ Closures for Thermo Scientific Nalgene Bottles (2135-5300)
  • Thermo Scientific Nalgene barbed bulkhead fitting kit, 1/2" tubing ID.

Headspace Vials 25 Ml

Buy Headspace Vials 25 Ml Items Including:
  • Cole-Parmer PTFE Vial, 25 mL (YO-06102-00)
  • 10 mL Headspace Vial with 20 mm Aluminum Seal Convenience Pack, PTFE/S (W225278-01)
  • Clear Precleaned EPA vials 60 mL case of 72 (YO-99535-25)
  • Thermo Scientific Nalgene Cryogenic vial; 1.0 mL, 25/pack.

Ami Instamatic Soil Ph Meter

Buy Ami Instamatic Soil Ph Meter Items Including:
  • Soil pH Electrode, BNC Connector (YO-05992-62)
  • Soil pH/conductivity meter kit (AG6/PH/PSTK)
  • Horiba Compact Nitrate Pocket Tester, for soil
  • Soil sampler extension rod, 26" long (PN004)
  • Replacement reagents for soil texture test kit (R-1067).

Bluelab Soil Ph Meter

Buy Bluelab Soil Ph Meter Items Including:
  • Soil pH Electrode, BNC Connector (YO-05992-62)
  • Soil pH/conductivity meter kit (AG6/PH/PSTK)
  • Horiba Compact Nitrate Pocket Tester, for soil
  • Soil sampler extension rod, 26" long (PN004)
  • Replacement reagents for soil texture test kit (R-1067).

Bluelab Truncheon Soil Ph Meter

Buy Bluelab Truncheon Soil Ph Meter Items Including:
  • Soil pH Electrode, BNC Connector (YO-05992-62)
  • Soil pH/conductivity meter kit (AG6/PH/PSTK)
  • Horiba Compact Nitrate Pocket Tester, for soil
  • Soil sampler extension rod, 26" long (PN004)
  • Replacement reagents for soil texture test kit (R-1067).

Farmtek Soil Ph Meter

Buy Farmtek Soil Ph Meter Items Including:
  • Soil pH Electrode, BNC Connector (YO-05992-62)
  • Soil pH/conductivity meter kit (AG6/PH/PSTK)
  • Horiba Compact Nitrate Pocket Tester, for soil
  • Soil sampler extension rod, 26" long (PN004)
  • Replacement reagents for soil texture test kit (R-1067).

Instamatic Ph Soil Ph Meter

Buy Instamatic Ph Soil Ph Meter Items Including:
  • Soil pH/conductivity meter kit (AG6/PH/PSTK)
  • Soil pH Electrode, BNC Connector (YO-05992-62)
  • Horiba Compact Nitrate Pocket Tester, for soil
  • Soil sampler extension rod, 26" long (PN004)
  • Replacement reagents for soil texture test kit (R-1067).

Instamatic Soil Ph Meter

Buy Instamatic Soil Ph Meter Items Including:
  • Soil pH Electrode, BNC Connector (YO-05992-62)
  • Soil pH/conductivity meter kit (AG6/PH/PSTK)
  • Horiba Compact Nitrate Pocket Tester, for soil
  • Soil sampler extension rod, 26" long (PN004)
  • Replacement reagents for soil texture test kit (R-1067).

Kelway Soil Ph Meter

Buy Kelway Soil Ph Meter Items Including:
  • Soil pH Electrode, BNC Connector (YO-05992-62)
  • Soil pH/conductivity meter kit (AG6/PH/PSTK)
  • Horiba Compact Nitrate Pocket Tester, for soil
  • Soil sampler extension rod, 26" long (PN004)
  • Replacement reagents for soil texture test kit (R-1067).



Love Temperature Controller Vendors to Kelway Soil Ph Meter

>