Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 5
Stainless Steel Sanitary Fittings
to
Thermo Scientific Nalgene Autoclavable Trays
Image
Product Detail
More Detail
Stainless Steel Sanitary Fittings
Buy Stainless Steel Sanitary Fittings Items Including:
Stainless steel sanitary bottle; 1 liter, 2" flange (PS-10F)
Stainless steel sanitary bottle; 10 liter, 4" flange (BTB-21)
Stainless steel sanitary bottle; 20 liter, 4" flange (BTB-27)
Stainless Steel Sanitary Check Valve, 1-½".
More Details
Cole-Parmer Asme-Bpe Biopharm Sanitary Clamp Fittings
Buy Cole-Parmer Asme-Bpe Biopharm Sanitary Clamp Fittings Items Including:
316L Stainless Steel Asme-bpe Sanitary Clamp 90° Elbow, 20RA Mecha (YO-30606-01)
316L Stainless Steel Asme-bpe Sanitary Clamp Concentric Reducer, 15ra (YO-30607-40)
316L Stainless Steel Asme-bpe Sanitary Clamp Concentric Reducer, 15ra (YO-30607-41).
More Details
Microscope/Telescopes with Built-in Reticles
Buy Microscope/Telescopes with Built-in Reticles Items Including:
Microscope/telescopes with built-in reticle; magnification, 25x/8x; fi (M820-S)
Microscope/telescopes with built-in reticle; magnification, 25x/8x; fi (M830-S)
Microscope/telescopes with built-in reticle; magnification, 25x/8x; fi (M820-SM).
More Details
Luxo Accessories for Circular Illuminated Magnifiers
Buy Luxo Accessories for Circular Illuminated Magnifiers Items Including:
10-Diopter Swing-away Auxiliary Lens.
More Details
Gast Vacuum Pumps
Buy Gast Vacuum Pumps Items Including:
Vacuum/pressure diaphragm pump with gauges, regulators, and relief val (DAA-V515-ED)
Vacuum/pressure diaphragm pump, single head, 1.1 cfm, 220/240VAC (DOA-P725-BN)
Vacuum/pressure diaphragm pump with gauges, regulators, and relief val (DOA-P704-AA).
More Details
Advantec Nobuto Filter Strips
Buy Advantec Nobuto Filter Strips Items Including:
Advantec Nobuto Blood Filter Strip, 100/pk.
More Details
Ceramic Hot Plates
Buy Ceramic Hot Plates Items Including:
Thermo Scientific Super-Nuova™ ceramic hot plate, 7 x 7"
EchoTherm™ Digital Hot Plate, Ceramic, 115 VAC
EchoTherm™ Digital Hot Plate, Ceramic, 230 VAC
Corning Digital Hot Plate, 5" x 7", 120 VAC
Economical Hot Plate, 120 VAC, 60 Hz.
More Details
Gate Valves
Buy Gate Valves Items Including:
Bronze gate valve; 1" NPT(F) (GV0700150)
Bronze gate valve; 1-1/4" NPT(F) (GV0700160).
More Details
Chemical Transfer Pumps
Buy Chemical Transfer Pumps Items Including:
Chemical-Duty Transfer Pump with Carbon-Filled Kynar Head, 38 GPM max
Chemical-Duty Transfer Pump with Carbon-Filled Kynar Head, 53 GPM max
Chemical-Duty Transfer Pump with Glass-Filled Polypropylene Head, Self.
More Details
Solenoid Pinch Valve
Buy Solenoid Pinch Valve Items Including:
Three-way solenoid pinch valve; 12 VDC, 1/8" ID x 1/4" OD tubing (100P3MP-12-05)
Three-way solenoid pinch valve; 24 VDC, 1/8" ID x 1/4" OD tubing (100P3MP-24-05).
More Details
Plastic Quick Connect Fittings
Buy Plastic Quick Connect Fittings Items Including:
Quick-disconnect fittings, Polysulfone fittings, 1/4" ID, body, 5/pack (MPC17004T39)
Quick-disconnect fittings, Polysulfone fittings, 3/8" ID, body, 5/pack (MPC17006T39).
More Details
Well Water Testing Kits
Buy Well Water Testing Kits Items Including:
Watersafe Bacteria Test Kit
Watersafe Pesticide Test Kit
Microbial Test Kits, Yeast and Mold Test Kit (120)
CHEMets Colorimetric Dissolved Oxygen Test Kit, 0 to 40 ppb (K-7540)
CHEMets Colorimetric Dissolved Oxygen Test Kit, 0 to 100 ppb (K-7599).
More Details
Chart Recorder Paper Houston
Buy Chart Recorder Paper Houston Items Including:
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C304)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C312)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C320).
More Details
Chart Recorder Paper Jewett
Buy Chart Recorder Paper Jewett Items Including:
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C304)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C312)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C320).
More Details
Chart Recorder Paper Supplies
Buy Chart Recorder Paper Supplies Items Including:
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C304)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C312)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C320).
More Details
Cobex Chart Recorder Paper
Buy Cobex Chart Recorder Paper Items Including:
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C304)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C312)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C320).
More Details
Custom Chart Recorder Paper
Buy Custom Chart Recorder Paper Items Including:
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C304)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C312)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C320).
More Details
Fisher Chart Recorder Paper
Buy Fisher Chart Recorder Paper Items Including:
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C304)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C312)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C320).
More Details
Gould Chart Recorder Paper
Buy Gould Chart Recorder Paper Items Including:
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C304)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C312)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C320).
More Details
Mb Chart Recorder Paper
Buy Mb Chart Recorder Paper Items Including:
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C304)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C312)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C320).
More Details
Buchi Rotary Evaporator Company
Buy Buchi Rotary Evaporator Company Items Including:
100-mL evaporator flask for Buchi Rotary Evaporators, 24/40 joint (YO-23123-62)
Heating bath for Buchi Rotary Evaporators, holds up to 5L flask, 115V (YO-23123-52)
Heating bath for Buchi Rotary Evaporators, holds up to 5L flask, 230VA (YO-23123-53).
More Details
Buchi Rotary Evaporator Models
Buy Buchi Rotary Evaporator Models Items Including:
100-mL evaporator flask for Buchi Rotary Evaporators, 24/40 joint (YO-23123-62)
Heating bath for Buchi Rotary Evaporators, holds up to 5L flask, 115V (YO-23123-52)
Heating bath for Buchi Rotary Evaporators, holds up to 5L flask, 230VA (YO-23123-53).
More Details
Buchi Rotary Evaporator R 210
Buy Buchi Rotary Evaporator R 210 Items Including:
Rotary Evaporator, Diagonal Condenser, Advanced Controller, 220V (YO-23012-01)
Rotary Evaporator, Safety-Coated Diagonal Condenser, Advanced Controll (YO-23012-06)
Rotary Evaporator, Safety-coated Diagonal Condenser, Professional Cont (YO-23012-07).
More Details
Buchi Rotary Evaporator Rotovap Rent
Buy Buchi Rotary Evaporator Rotovap Rent Items Including:
100-mL evaporator flask for Buchi Rotary Evaporators, 24/40 joint (YO-23123-62)
Heating bath for Buchi Rotary Evaporators, holds up to 5L flask, 115V (YO-23123-52)
Heating bath for Buchi Rotary Evaporators, holds up to 5L flask, 230VA (YO-23123-53).
More Details
Replacement Buchi Rotary Evaporator Flange
Buy Replacement Buchi Rotary Evaporator Flange Items Including:
100-mL evaporator flask for Buchi Rotary Evaporators, 24/40 joint (YO-23123-62)
Heating bath for Buchi Rotary Evaporators, holds up to 5L flask, 115V (YO-23123-52)
Heating bath for Buchi Rotary Evaporators, holds up to 5L flask, 230VA (YO-23123-53).
More Details
Stir Bars
Buy Stir Bars Items Including:
Large-Volume Stir Bar, 4-1/2"L x 1-1/16"dia (YO-04627-00)
Scienceware Spinbar Smco, Stirring Bar, PTFE, 2" X 7/8"
Scienceware Spinvane Stirring Bar, PTFE, Triangular, 1mL
Scienceware Spinvane Stirring Bar, PTFE, Triangular, 3mL.
More Details
Thermo Scientific Orion Ph Electrode
Buy Thermo Scientific Orion Ph Electrode Items Including:
Thermo Scientific Orion pH electrode, glass body, refillable, single-j (9102BNWP)
Thermo Scientific Orion pH electrode, glass body, refillable, single-j (9103BNWP)
Thermo Scientific Orion pH electrode, glass body, refillable, single-j (9104BNWP).
More Details
Gilmont Falling Ball Viscometer
Buy Gilmont Falling Ball Viscometer Items Including:
Gilmont Falling Ball Viscometer, 20 to 200 / 100 to 1000 centipoise
Gilmont Falling Ball Visometer, 2 to 20 / 10 to 100 centipoise
Gilmont Fallng Ball Viscometer, 0.2 to 2 / 1 to 10 centipoise
Gas Ball for Falling Ball Viscometer (8000009).
More Details
Pyrex Flasks
Buy Pyrex Flasks Items Including:
PYREX Brand 4980 flask; 125 mL, pack of 12
PYREX Brand 4980 flask; 250 mL, pack of 12
PYREX Brand 4980 flask; 300 mL, pack of 12
PYREX Brand 4980 flask; 1000 mL, pack of 6
PYREX Brand 4980 flask; 2000 mL, pack of 1.
More Details
Thermo Scientific Nalgene Autoclavable Trays
Buy Thermo Scientific Nalgene Autoclavable Trays Items Including:
Thermo Scientific Nalgene polypropylene autoclaving tray, 1 L
Thermo Scientific Nalgene polypropylene autoclaving tray, 2 L
Thermo Scientific Nalgene polypropylene autoclaving tray, 3 L
Thermo Scientific Nalgene polypropylene autoclaving tray, 5 L.
More Details
70618
results
<<<Previous
1
2
3
4
5
6
7
8
9
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Stainless Steel Sanitary Fittings to Thermo Scientific Nalgene Autoclavable Trays
>
31125329-s-toc_2 31125329stoc2