Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 6
Thermo Scientific Nalgene Cryobox Storage Boxes
to
Tank Spray Guns
Image
Product Detail
More Detail
Thermo Scientific Nalgene Cryobox Storage Boxes
Buy Thermo Scientific Nalgene Cryobox Storage Boxes Items Including:
Thermo Scientific Nalgene Polycarbonate CryoBox storage boxes for 25 (
Thermo Scientific Nalgene Polycarbonate CryoBox storage boxes for 81 ( (5026-0909)
Thermo Scientific Nalgene Polycarbonate CryoBox storage boxes for 81 ( (5027-0909).
More Details
Flaring Tools for Pfa Flare Fittings
Buy Flaring Tools for Pfa Flare Fittings Items Including:
Flaring tool for tubing 1/4 to 1" outer diameter (YO-30490-01)
Flaring tool for tubing 1/4" outer diameter (YO-30490-03)
Flaring tool for tubing 3/8" outer diameter (YO-30490-05).
More Details
Sartorius Analytical Balance
Buy Sartorius Analytical Balance Items Including:
Sartorius Extend™ ED Analytical Balance, 120 g x 0.1 mg
Practum Analytical Balance, 220g by 0.1mg, External Calibration
Practum Analytical Balance, 120g by 0.1mg, External Calibration
Quintix Analytical Balance, 220g by 0.1mg, Internal Calibration.
More Details
Weighing Paper
Buy Weighing Paper Items Including:
Glassine Weighing Paper 3x3" 500/pk (YO-01338-00)
Glassine Weighing Paper 4x4" 500/pk (YO-01338-02)
Glassine Weighing Paper 6x6" 500/pk (YO-01338-04).
More Details
Parker Plastic Check Valves
Buy Parker Plastic Check Valves Items Including:
CHECK VALVE, ACETAL BODY, push-to-connect, 1/4'' tube OD (A4VC4-MG)
CHECK VALVE, ACETAL BODY, push-to connect, 5/16'' tube OD (A5VC5-MG).
More Details
Cole-Parmer Nylon Monofilament Filter Bags
Buy Cole-Parmer Nylon Monofilament Filter Bags Items Including:
Nylon monofilament filter bag; 10"L, 100 µm (255009-03)
Nylon monofilament filter bag; 10"L, 200 µm (255011-03)
Nylon monofilament filter bag; 10"L, 250 µm (BN-410-250)
Nylon monofilament filter bag; 18-1/2"L, 50 µm (255255-03).
More Details
Unf Female Threaded Quick-Disconnect Coupling Inserts
Buy Unf Female Threaded Quick-Disconnect Coupling Inserts Items Including:
Metal valved quick-disconnect female UNF 10-32 threaded insert, 1/8" f (MCD281032).
More Details
Metric Compression Fitting
Buy Metric Compression Fitting Items Including:
Compression To Thread Adapter, Stainless Steel, 6mm OD Tubing X 1/4" N (YO-31528-02)
Compression Straight Union, Stainless Steel, 6mm OD Tubing (YO-31528-33)
Compression Straight Union, Stainless Steel, 8mm OD Tubing (YO-31528-34).
More Details
Bellows Metering Pumps
Buy Bellows Metering Pumps Items Including:
Bellows metering pump, 6 to 30 mL/min, 115 VAC, 50/60 Hz (14280-003-X006-T001)
Bellows metering pump with Viton valve and O-ring, 30 to 300 mL/min, 1
Bellows metering pump with Viton valve and O-ring, 173 to 1730 mL/min, (14251-007-X116-T08).
More Details
True Union Ball Check Valves
Buy True Union Ball Check Valves Items Including:
Hayward True Union Ball Check Valve, 1/2" socket and NPT(F) connectors
Hayward True Union Ball Check Valve, 3/4" socket and NPT(F) connectors
Hayward True Union Ball Check Valve, 1-1/2" socket and NPT(F) connecto.
More Details
Polypropylene Spectra/Mesh Screen Discs and Sheets
Buy Polypropylene Spectra/Mesh Screen Discs and Sheets Items Including:
Spectra/Mesh Screen Discs, Polypropylene, 47 mm, 105 µm, pack of 10
Spectra/Mesh Screen Discs, Polypropylene, 47 mm, 149 µm, pack of 10
Spectra/Mesh Screen Discs, Polypropylene, 47 mm, 210 µm, pack of 10.
More Details
Threaded Plugs
Buy Threaded Plugs Items Including:
Threaded plug, nylon, 1" male NPT, 10/pack (YO-30401-36)
Threaded plug, PFA, 1/8" male NPT (YO-31321-13)
Plug Valve with Pipe Thread Connections, 1/4" NPT(F) (D1071794)
Tube end plug, PTFE/Tefzel, for 1/4"-28 UNF thread, 2/pack.
More Details
Stainless Luer Fittings
Buy Stainless Luer Fittings Items Including:
316 Stainless Steel fittings, female luer x female luer (YO-31507-89)
316 Stainless Steel fittings, male luer lock x male luer lock (YO-31507-90)
Female luer lock x 1/4" NPT, 316 SS, each (YO-31507-48).
More Details
Gaskets for Ansi 150# Flanges
Buy Gaskets for Ansi 150# Flanges Items Including:
ANSI 150# Ff Gasket,epdm,1/2" (YO-30549-01).
More Details
John Guest Push-to-Connect Fittings
Buy John Guest Push-to-Connect Fittings Items Including:
John Guest Push-to-Connect Collet Cover, 5/16" Tube OD
Push-to-connect soft tube supports, 1/2" stem x 3/8" tube OD, 10/pack (YO-34007-39)
Push-to-connect soft tube supports, 5/16" stem x 1/4" tube OD, 10/pack (YO-34007-40).
More Details
Water Test Strips for Bacteria
Buy Water Test Strips for Bacteria Items Including:
LaMotte Alkalinity Test Strips, 0 to 180 ppm
LaMotte Free Chlorine Test Strips, 0 to 10 ppm
LaMotte Total Chlorine Test Strips, 0 to10 ppm
LaMotte Total Chlorine Test Strips, 0 to 5 ppm
EM Quant Ammonium Test Strips
More Details
Glass Stirring Rods
Buy Glass Stirring Rods Items Including:
Cole-Parmer Glass Stir Rods with Rubber Policeman, 10" 12/pk (YO-06369-56)
Cole-Parmer Glass Stir Rods with Rubber Policeman, 12" 12/pk (YO-06369-58)
Stir Rod, Glass, 150mm, 200/pk (40500-150)
Stir Rod, Glass, 250mm, 100/pk (40500-250).
More Details
Oil-Less Air Compressor, Piston Compressor Pumps
Buy Oil-Less Air Compressor, Piston Compressor Pumps Items Including:
Oilless Air Compressor, Piston compressor pump, 3.5 cfm, 115/230 VAC (4HCC-10-M450X)
Oilless Air Compressor, Piston compressor pump, 9.1 cfm, 220/380 VAC (7HDD-10-M853)
Oilless Air Compressor, Piston compressor pump, 11 cfm, 230 VAC (8HDM-10-M850X).
More Details
Bradley Combination Showers
Buy Bradley Combination Showers Items Including:
Bradley Combination Showers-Shower/eyewash station
Bradley Combination Showers-Shower/eye/facewash station
Bradley Combination Showers (foot operated).
More Details
Ysi Conductivity Calibration Solutions
Buy Ysi Conductivity Calibration Solutions Items Including:
YSI conductivity calibration standard, 10,000 µS
YSI conductivity calibration standard, 100,000 µS.
More Details
Plastic Siphon Drum Pump
Buy Plastic Siphon Drum Pump Items Including:
Hand-operated water and chemical siphon/drum pump, 32 strokes/gallon
Hand-operated chemical siphon/drum pump, 12 strokes/gallon
Hand-operated siphon/lift drum pump, 16 strokes/gallon (MP1848S).
More Details
Polypropylene Bag Filter Housings
Buy Polypropylene Bag Filter Housings Items Including:
Nylon monofilament filter bag; 10"L, 100 µm (255009-03)
Nylon monofilament filter bag; 10"L, 200 µm (255011-03)
Nylon monofilament filter bag; 10"L, 250 µm (BN-410-250)
Nylon monofilament filter bag; 10"L, 300 µm (255013-03).
More Details
Troemner Gas Cylinder Stands
Buy Troemner Gas Cylinder Stands Items Including:
Stand (970040)
Portable Stand
PVC Coated Stand.
More Details
Stainless Steel Water Tank
Buy Stainless Steel Water Tank Items Including:
Stainless steel cylindrical utility tank with cover, 18-18 stainless s (YO-07233-10)
Stainless steel cylindrical utility tank with cover, 18-8 stainless st (YO-07233-20)
Stainless steel cylindrical utility tank with cover, 2 7/8 gal
(YO-07228-00).
More Details
Bulkhead Mounted 2 Plastic Ball Valve
Buy Bulkhead Mounted 2 Plastic Ball Valve Items Including:
Ball Valve, 2-way, 1/2" barb, PP with EPDM seals (YO-01377-54)
Ball Valve, 2-way, 1/2" barb, PP with Viton® seals
Ball Valve, 2-way, 1/8" NPT(F), PP with EPDM seals
Ball Valve, 2-way, 1/4" barb, PP with EPDM seals (YO-01377-34).
More Details
Pyrex Graduated Cylinder
Buy Pyrex Graduated Cylinder Items Including:
Pyrex 3022 Cylinder, 10ml, 24/cs
Pyrex 3022 Cylinder, 25ml, 18/cs
Pyrex 3022 Cylinder, 50 ml, 18/cs
Pyrex 3022 Cylinder, 100 ml, 12/cs
Pyrex 3022 Cylinder, 250ml, 12/cs
More Details
High-Efficiency Axial Flow Impellers (Lightnin a-310)
Buy High-Efficiency Axial Flow Impellers (Lightnin a-310) Items Including:
Lightnin® (A-310) 316 SS high-efficiency axial flow impeller, 3.8" (871581PSP)
Lightnin® (A-310) 316 SS high-efficiency axial flow impeller, 4.5" (808887PSP)
Lightnin® (A-310) 316 SS high-efficiency axial flow impeller, 7.6" (871235).
More Details
Hach Turbidity Standards and Calibration Sets
Buy Hach Turbidity Standards and Calibration Sets Items Including:
Hach StablCal 0.1 NTU Turbidity Standard, 500-mL
Hach StablCal 20 NTU Turbidity Standard, 500-mL
Turb Standard 0.0, Hach 2100q
Turb Standard 20, Hach 2100q
Turb Standard 100, Hach 2100q
More Details
Yamato Programmable Mechanical Convection Ovens
Buy Yamato Programmable Mechanical Convection Ovens Items Including:
Yamato Programmable Mechanical Convection Oven, 3.2 Cu Ft, 115VAC (DKN-402)
Yamato Programmable Mechanical Convection Oven, 5.3 Cu Ft, 115VAC (DKN-602)
Yamato Programmable Mechanical Convection Oven, 10.7 Cu Ft, 220 VAC.
More Details
Tank Spray Guns
Buy Tank Spray Guns Items Including:
Spray Gun PTFE Di H20 3/8" (THG-DN6P-TFEV)
Spray Gun PTFE Di H20 1/2" (THG-DN8P-TFEV)
Spray Gun Recirculating 3/8" (THG-DR6P-TFE)
Spray Gun Recirculating Assmby (THG-DR6PVHE02)
Dispensing Gun PTFE 3/8"npt (THG-LD6P-TFEV).
More Details
70618
results
<<<Previous
2
3
4
5
6
7
8
9
10
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Thermo Scientific Nalgene Cryobox Storage Boxes to Tank Spray Guns
>
31125329-s-toc_2 31125329stoc2