Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 60
Surgical Rubber Tubing 1 2 Id
to
Ph Buffers
Image
Product Detail
More Detail
Surgical Rubber Tubing 1 2 Id
Buy Surgical Rubber Tubing 1 2 Id Items Including:
Black Rubber Vacuum Tubing, 1/2" ID x 1 1/8" OD, 25ft
(YO-96143-12)
Black Rubber Vacuum Tubing, 1/2" ID x 1 1/8" OD, 25ft
(YO-96143-13)
Black Rubber Vacuum Tubing, 1/4" ID x 1/2" OD, 25ft
More Details
Soil Sieve Set Stainless Steel
Buy Soil Sieve Set Stainless Steel Items Including:
Receiving Pan for 12" Stainless Steel Bottom Sieves, Full Height
Receiving Pan with Fitted Rim for Nesting 12" Stainless Steel Sieves, (YO-59996-92)
Receiving Pan with Fitted Rim for Nesting 12" Stainless Steel Sieves, (YO-59996-93).
More Details
Advantec Grade Gb140 Glass Fiber Filters
Buy Advantec Grade Gb140 Glass Fiber Filters Items Including:
Advantec Grade GB140 Glass Fiber Filters; 47mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 90mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 125mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 142mm dia; 100/box.
More Details
Miltex Needle Holders
Buy Miltex Needle Holders Items Including:
Olsen-Hegar Needle Holders, Dissecting-grade 5.5" (YO-10921-74)
Olsen-Hegar Needle Holders, Dissecting-grade 7.5" (YO-10921-76)
Mayo-Hegar Needle Holders, Precision-grade (YO-10921-78)
Olsen-Hegar Needle Holders, Precision-grade 5".
More Details
Accumet Ph Electrodes Refilling
Buy Accumet Ph Electrodes Refilling Items Including:
Accumet pH Electrode, Combination, Microprobe, Refillable, Glass Body,
Accumet Accuphast pH Electrode, Combination, Microprobe, Refillable, G
Accumet Accutuph Plus pH Electrode, Combination, Refillable, Glass Bod.
More Details
Scienceware Spinbar Ptfe Stirring Bars
Buy Scienceware Spinbar Ptfe Stirring Bars Items Including:
Scienceware SPINBAR PTFE ROUND 1" x 5/16"
Scienceware Spinbar Smco, Stirring Bar, PTFE, 2" X 7/8"
Scienceware Spinbar Stirring Bar, PTFE, Flea Micro, Blue, 6.35x3mm
Scienceware Spinbar Stirring Bar, PTFE, Flea Micro, Red, 7x2mm.
More Details
Pharmafluor Pfa Compression Union Elbows
Buy Pharmafluor Pfa Compression Union Elbows Items Including:
Pharmafluor Straight Compression Elbow, PFA, 1/4 (YO-31001-12).
More Details
Pvdf Tee Fittings
Buy Pvdf Tee Fittings Items Including:
Installation Fittings, tee pvdf for 1/2" ID pipe (SFMT005)
Installation Fittings, tee pvdf for 1-1/4" ID pipe (SFMT012)
PVDF Sanitary Tee, 3/4" Mini Clamp Size (YO-31809-61)
PVDF Sanitary Tee, 1" Ladish Clamp Size (YO-31809-62).
More Details
Cellulose Acetate Membrane
Buy Cellulose Acetate Membrane Items Including:
Corning Disposable Bottletop Filter; 150-mL volume; CA, membrane; 48/p
Corning Disposable Bottletop Filter; 500-mL volume; CA, membrane; 12/p
Corning Disposable Bottletop Filter; 1000-mL volume; CA, membrane; 12/.
More Details
Vantage Pro2 Weather Station
Buy Vantage Pro2 Weather Station Items Including:
Davis Instruments Vantage Pro2 Plus Cabled Weather Station System
Davis Instruments Vantage Pro2™ Plus Wireless Weather Station with
Davis Instruments Vantage Pro2™ Wireless Integrated Sensor Suite
Davis Instruments 6625 Vantage Pro2 Power Adapter.
More Details
Filters for 30 Inch Fume Hoods
Buy Filters for 30 Inch Fume Hoods Items Including:
Primary Hydrocarbon Filters for 30" Self-Monitoring Ductless Fume Hood (YO-33713-50)
Secondary Hydrocarbon Filters for 30" Self-Monitoring Ductless Fume Ho (YO-33713-55).
More Details
Hdpe Tote Boxes
Buy Hdpe Tote Boxes Items Including:
HDPE tote box, 0.5 cu ft (35-180 BLUE)
HDPE tote box, 1.2 cu ft (35-190 BLUE)
HDPE tote box, 2.6 cu ft (35-230 BLUE)
HDPE tote box, 3.7 cu ft (35-300 BLUE)
Box, HDPE, 2 gallon
More Details
Polycarbonate Luer Lock Ring
Buy Polycarbonate Luer Lock Ring Items Including:
Snap luer lock ring, PC, 25/pk (YO-45504-74)
Snap luer lock ring, Nylon, 25/pk (YO-45505-74)
Male luer with lock ring x 1/16" hose barb, PC, 25/pk (YO-45504-00)
Male luer with lock ring x 3/32" hose barb, PC, 25/pk (YO-45504-02).
More Details
Rubber Coated Gloves
Buy Rubber Coated Gloves Items Including:
Sperian® Atlas Fit™ Rubber Coated Cotton/Poly Glove, Medium
Atlas Fit® Rubber Coated Cotton Poly Glove Black and Gray Sm (320-S).
More Details
Cotton Lined Rubber Gloves
Buy Cotton Lined Rubber Gloves Items Including:
MAPA Butoflex 650 Cotton-Lined Butyl Gloves, size 7, 1 pair
MAPA Butoflex 650 Cotton-Lined Butyl Gloves, size 8, 1 pair
MAPA Butoflex 650 Cotton-Lined Butyl Gloves, size 9, 1 pair
MAPA Butoflex 650 Cotton-Lined Butyl Gloves, size 10, 1 pair.
More Details
Ace Glass Centrifuge Tubes with Filter
Buy Ace Glass Centrifuge Tubes with Filter Items Including:
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 5 ml, 125 (99502-5)
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 10 ml, 12 (99502-10)
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 15 ml, 12 (99502-15).
More Details
Oil Free Diaphragm Vacuum Pump
Buy Oil Free Diaphragm Vacuum Pump Items Including:
Vacuum Pump, Diaphragm Vacuum Pump - 1.23 CFM, 2 Stage, 115 VAC, 50/60
Vacuum Pump, Brandtech Vacuum Pump - 1.4 CFM, 120 VAC, 50/60 Hz, 2 Sta (732003)
Vacuum Pump, Brandtech Vacuum Pump - 1.4 CFM, 230 VAC, 50 Hz, 2 Stage, (732000).
More Details
Micro Scale Magnetic Stir Bar
Buy Micro Scale Magnetic Stir Bar Items Including:
Spinbar Magnetic Stir Bar Rack
Glass Encased magnetic Stir Bars, 1/2"l X 3/8"d (F37101-0012)
Micro Stir Bar, 7mm L x 2mm dia (YO-08545-00)
Micro Stir Bar, 6.35mm L x 3mm dia (YO-08545-02)
Micro Stir Bar, 12.7mm L x 3mm dia (YO-08545-03).
More Details
Black Latex Glove
Buy Black Latex Glove Items Including:
Microflex Black Dragon Latex Gloves, X-small, 100/box
Microflex Black Dragon Latex Gloves, Small, 100/box
Microflex Black Dragon Latex Gloves, Medium, 100/box
Microflex Black Dragon Latex Gloves, Large, 100/box.
More Details
John Guest Acetal Push-to-Connect Straight Unions
Buy John Guest Acetal Push-to-Connect Straight Unions Items Including:
Straight Unions, 1/4, 10 Per Pack (YO-34006-17)
Straight Unions, 3/8, 10 Per Pack (YO-34006-18)
Straight Unions, 1/2, 10 Per Pack (YO-34006-19)
Union Connector, 3/16, 10 Per Case (YO-34006-32)
Union Connector, 5/32, 10 Per Case (YO-34006-57).
More Details
Kimble-Chase Measuring (Mohr) Pipettes
Buy Kimble-Chase Measuring (Mohr) Pipettes Items Including:
Kimble-Chase Long-Tip Measuring (Mohr) Pipettes, 1 mL
Kimble-Chase Long-Tip Measuring (Mohr) Pipettes, 2 mL
Kimble-Chase Long-Tip Measuring (Mohr) Pipettes, 5 mL.
More Details
Polypropylene Sanitary Clamp Concentric Reducers
Buy Polypropylene Sanitary Clamp Concentric Reducers Items Including:
Polypropylene Sanitary Concentric Reducer,1.5X3/4" (YO-31805-06)
Polypropylene Sanitary Concentric Reducer, 2" x 1" Ladish Clamp Size (YO-31805-11)
Polypropylene Sanitary Concentric Reducer, 3" x 2" Clamp Size (YO-31805-18).
More Details
Thermo Scientific Labquake Shakers
Buy Thermo Scientific Labquake Shakers Items Including:
Thermo Scientific Labquake Tube Shaker/Rotators, Large, 14 Tubes, 50 m (4152110Q)
Thermo Scientific Labquake Tube Shaker/Rotators, Large, 14 Tubes, 50 m (4152220Q)
Thermo Scientific Labquake Tube Shaker/Rotators, Large, 46 Tubes, 10 m (T415110Q).
More Details
4 Butterfly Valve Dwg
Buy 4 Butterfly Valve Dwg Items Including:
Sanitary Butterfly Valve, 4" TRI-CLAMP.
More Details
Thin Glass Microscope Slides
Buy Thin Glass Microscope Slides Items Including:
Superfrost Excell Adhesion Microscope Slides, 144/pk
Superfrost Excell Clipped Corner Adhesion Microscope Slides, 144/pk
Cole-Parmer Brand Slides; blue frosted; one gross (144 slides)
Cole-Parmer Brand Slides; green frosted; one gross (144 slides).
More Details
Metal G Thread Fittings
Buy Metal G Thread Fittings Items Including:
Metal quick-disconnect coupling body, valved, 1/4" NPT (M) thread, 1/4 (LCD10004)
Metal quick-disconnect coupling body, valved, 1/4" NPT (M) thread, 3/8 (LCD10006).
More Details
Glass Vial with Measuring Cap
Buy Glass Vial with Measuring Cap Items Including:
Glass Vials PTFE Lined Cap Clear 12 ml 200/pk (B7800-12)
Glass Vials PTFE Lined Cap Clear 16 ml 200/pk (B7800-4)
Glass Vials PTFE Lined Cap Clear 20 ml 100/pk (B7800-20)
Glass Vials PTFE Lined Cap Clear 22 ml 200/pk (B7800-5).
More Details
Ansell Latex Gloves
Buy Ansell Latex Gloves Items Including:
Ansell Reusable unlined natural latex gloves; 20-mil thick; 12"L; size
Conform XT Latex Gloves, Small, 100/bx
Conform XT Latex Gloves, Large, 100/bx
Conform XT Latex Gloves, X-Large, 100/bx
Ansell AccuTech Sterile Latex Gloves, Size 7, 20/BX.
More Details
47 Mm Glass Filter Paper
Buy 47 Mm Glass Filter Paper Items Including:
Whatman Prepleated Filter Papers; 24 cm dia; pore size 8 µ; 100/box
Whatman Glass Microfiber Filter Papers Water-Pollution test papers; 4
(YO-06648-70)
Whatman Glass Microfiber Filter Papers Water-Pollution test papers; 4
(YO-06648-71).
More Details
Ph Buffers
Buy Ph Buffers Items Including:
Thermo Scientific Orion PerpHecT pH buffer packs, 4.01, 10/PK
Thermo Scientific Orion PerpHecT pH buffer packs, 4.01, 25/PK
Thermo Scientific Orion PerpHecT pH buffer packs, 7.00, 10/PK
Thermo Scientific Orion PerpHecT pH buffer packs, 7.00, 25/PK.
More Details
70618
results
<<<Previous
56
57
58
59
60
61
62
63
64
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
20
30
40
45
46
47
48
49
50
51
52
53
54
55
56
57
58
59
60
61
62
63
64
65
66
67
68
69
70
71
72
73
74
80
90
100
110
120
130
140
150
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Surgical Rubber Tubing 1 2 Id to Ph Buffers
>
31125329-s-toc_2 31125329stoc2