Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 7

Allsafe Smc Nemesis Safety Glasses   to   Graduated Beakers with Handle

Image
  Product Detail
More Detail

Allsafe Smc Nemesis Safety Glasses

Buy Allsafe Smc Nemesis Safety Glasses Items Including:
  • Allsafe SMC™ Nemesis™ Safety Glasses, Black frame, Clear lens.

Paddle Blenders

Buy Paddle Blenders Items Including:
  • Paddle Blender; 50 To 400 mL, Variable Speed, Window Door, Removable P (YO-36850-13)
  • Paddle Blender; 200 To 3500 mL, Variable Speed, Stainless Steel Door
  • PADDLE BLENDER STARTER KIT; 400 ML Fixed speed blender with transparen (YO-36850-17).

Advantec 47 Mm Glass Fiber Filters

Buy Advantec 47 Mm Glass Fiber Filters Items Including:
  • Advantec Grade GA55 Glass Fiber Filters, 125mm dia; 100/box
  • Advantec Grade GB140 Glass Fiber Filters; 47mm dia; 100/box
  • Advantec Grade GB140 Glass Fiber Filters; 90mm dia; 100/box
  • Advantec Grade GB140 Glass Fiber Filters; 125mm dia; 100/box.

Advantec Glass Fiber Filter Sheets

Buy Advantec Glass Fiber Filter Sheets Items Including:
  • Advantec Grade GA55 Glass Fiber Filters, 125mm dia; 100/box
  • Advantec Grade GB140 Glass Fiber Filters; 47mm dia; 100/box
  • Advantec Grade GB140 Glass Fiber Filters; 90mm dia; 100/box
  • Advantec Grade GB140 Glass Fiber Filters; 125mm dia; 100/box.

Pulsafeeder Metering Pumps

Buy Pulsafeeder Metering Pumps Items Including:
  • Service Kit for Solenoid Metering Pump, 240 GPD
  • 4 to 20 mA pace pumps, 12 GPD, 115 VAC
  • 4 to 20 mA pace pumps, 12 GPD, 230 VAC
  • 4 to 20 mA pace pumps, 21 GPD, 115 VAC
  • 4 to 20 mA pace pumps, 21 GPD, 230 VAC

Slide Warmer

Buy Slide Warmer Items Including:
  • Slide warmer, 115 VAC 50/60 Hz (YO-48573-01).

Nylon Monofilament Filter Bags

Buy Nylon Monofilament Filter Bags Items Including:
  • Size 1 high-flow nylon monofilament filter bag; 300°F (149°C) (255052-03)
  • Size 1 high-flow nylon monofilament filter bag; 300°F (149°C) (255056-03)
  • Size 2 high-flow nylon monofilament filter bag; 300°F (149°C) (255068-03).

H-B Instrument Thermometer Storage Tubes

Buy H-B Instrument Thermometer Storage Tubes Items Including:
  • Plastic Tube Storage Case For 160mm Single Thermometer (20007)
  • Plastic Tube Storage Case For 305mm Single Thermometer (20008).

Chemets Colorimetric Dissolved Oxygen Test Kits

Buy Chemets Colorimetric Dissolved Oxygen Test Kits Items Including:
  • CHEMets Colorimetric Dissolved Oxygen Test Kit, 0 to 40 ppb (K-7540)
  • CHEMets Colorimetric Dissolved Oxygen Test Kit, 0 to 100 ppb (K-7599)
  • CHEMets Colorimetric Dissolved Oxygen Test Kit, 0 to 1 ppm (K-7501).

Corning Pyrex Brand Fiber Glass Wool

Buy Corning Pyrex Brand Fiber Glass Wool Items Including:
  • Fiber Glass Wool, 12-lb pack (3950).

Miniature Gear Pumps

Buy Miniature Gear Pumps Items Including:
  • Miniature gear pump, 0.43 GPM (1600 mL/min) non-enclosed OEM model, 11
  • Miniature gear pump, 0.43 GPM (1600 mL/min) non-enclosed OEM model, 23
  • Miniature gear pump, .32 GPM (1200 mL/min), fully enclosed lab model, (400000/LGP-115).

Latex Finger Cots

Buy Latex Finger Cots Items Including:
  • North Powdered Latex finger cots, small, 1440/bag
  • North Powdered Latex finger cots, medium, 1440/bag
  • North Powdered Latex finger cots, large, 1440/bag
  • QRP™ Latex Antistatic Finger Cots, small
  • QRP™ Latex Antistatic Finger Cots, medium.

Abb Commander Circular Chart Recorders

Buy Abb Commander Circular Chart Recorders Items Including:
  • ABB Commander Circular Chart Recorder/Controller - 1 controller, 2 pen
  • Chart Paper for ABB Commander Circular Chart Recorder/Controllers, 0 t (500P1225-106)
  • Chart Paper for ABB Commander Circular Chart Recorder/Controllers, 0 t (500P1225-15).

Uv Black Light Fluorescent

Buy Uv Black Light Fluorescent Items Including:
  • Black (UV) light ring lights, for 48402-series stereomicroscopes; 115V (YO-41210-00)
  • Black (UV) light ring lights, for 48403-series stereomicroscopes; 115V (YO-41210-10).

Laboratory Mortar and Pestle

Buy Laboratory Mortar and Pestle Items Including:
  • Cole-Parmer Mortar and Pestle Set, Porcelain; 80ML, Each
  • Cole-Parmer Mortar and Pestle Set, Porcelain; 500ML Each
  • Mortar and Pestle; Agate, 125ML, Each
  • Agate Mortar and Pestle, 25 mL (YO-04010-20)
  • Glass Mortar & Pestle Set, 50ml (YO-34500-51).

Pinnacle Ph/Ion Laboratory Electrodes

Buy Pinnacle Ph/Ion Laboratory Electrodes Items Including:
  • Three-in-one pH/reference/temperature electrode, epoxy body, BNC for m (476436P)
  • Replacement three-in-one pH electrode, gel-filled junction; 6-ft cable
  • Replacement high-performance pH electrode, combination, glass body, BN
  • ATC Probe (477968).

Accumet Glass Combination Electrodes

Buy Accumet Glass Combination Electrodes Items Including:
  • Accumet Accutuph pH Electrode, Combination, Single-junction, Refillabl
  • accumet accuTupH pH Electrode, Combination, Double-junction, Refillabl
  • Accumet pH Electrode, Combination, Microprobe, Refillable, Glass Body,.

Gas Cylinder Securing Safety Straps

Buy Gas Cylinder Securing Safety Straps Items Including:
  • Bench Clamp with secure message (973036)
  • Wall Bracket with secure message (973037)
  • Bench Clamp with secure message (974036)
  • Wall Bracket with secure message (974037)
  • Safety Strap without Message

Hayward Check Valves

Buy Hayward Check Valves Items Including:
  • Hayward True Union Ball Check Valve, 1/2" socket and NPT(F) connectors
  • Hayward True Union Ball Check Valve, 3/4" socket and NPT(F) connectors
  • Hayward True Union Ball Check Valve, 1-1/2" socket and NPT(F) connecto.

Water Testing Strips

Buy Water Testing Strips Items Including:
  • LaMotte Alkalinity Test Strips, 0 to 180 ppm
  • LaMotte Free Chlorine Test Strips, 0 to 10 ppm
  • LaMotte Total Chlorine Test Strips, 0 to10 ppm
  • LaMotte Total Chlorine Test Strips, 0 to 5 ppm
  • EM Quant Ammonium Test Strips

Metric Size Stainless Steel Compression Fittings

Buy Metric Size Stainless Steel Compression Fittings Items Including:
  • Compression To Thread Adapter, Stainless Steel, 6mm OD Tubing X 1/4" N (YO-31528-24)
  • Compression Straight Union, Stainless Steel, 10mm OD Tubing (YO-31528-35)
  • Compression Straight Union, Stainless Steel, 12mm OD Tubing (YO-31528-36).

Parker Brass Female/Female Manual Ball Valves

Buy Parker Brass Female/Female Manual Ball Valves Items Including:
  • 1" NPT brass female/female manual ball valves (XV500P-16)
  • 1/4" NPT brass female/male manual ball valves (XV501P-4)
  • 3/8" NPT brass female/male manual ball valves (XV501P-6)
  • 1" NPT brass female/male manual ball valves (XV501P-16).

Plastic Ball Valves

Buy Plastic Ball Valves Items Including:
  • George Fischer Type 546 Ball Valve, PVC with EPDM seals, 3/8" (161.546.341)
  • George Fischer Type 546 Ball Valve, PVC with EPDM seals, 1/2" (161.546.342)
  • George Fischer Type 546 Ball Valve, PVC with EPDM seals, 3/4" (161.546.343).

Magnifying Safety Glasses

Buy Magnifying Safety Glasses Items Including:
  • Olympic Optical magnifying safety glasses, black frame w/clear lens, + (OL30-06C1.5I)
  • Olympic Optical magnifying safety glasses, black frame w/clear lens, + (OL30-06C2.0I)
  • 3M Virtua™ 1.5 Magnifier Safety Eyewear, Clear Lens.

A&D Weighing Gemini Gr Analytical Balances

Buy A&D Weighing Gemini Gr Analytical Balances Items Including:
  • A&D Gemini Autocalibrating Analytical Balance, 120 G
  • A&D Gemini Autocalibrating Analytical Balance, 210 G.

Inch Compression Ferrule and Nut Assembly

Buy Inch Compression Ferrule and Nut Assembly Items Including:
  • Ferrule and nut assembly, stainless steel, 1/2" (YO-31212-63)
  • Ferrule and nut assembly, stainless steel, 3/8" (YO-31212-64)
  • Ferrule and nut assembly, brass, 1/8" (YO-31212-69)
  • Ferrule and nut assembly, brass, 1/4" (YO-31212-70).

Ph Buffer Tablets

Buy Ph Buffer Tablets Items Including:
  • Buffer tablets, pH 4, (bag of 50).

Sonicator Qsonica Ultrasonic Processors

Buy Sonicator Qsonica Ultrasonic Processors Items Including:
  • Open end wrench (requires 2) for use with ultrasonic processors
  • Replacement tube holder (20 tubes) for use with ultrasonic processors
  • Dual Horn (without probes) for use with ultrasonic processors
  • Stand with Clamp for use with ultrasonic processors.

Shimadzu Uv-1800 Uv/Visible Scanning Spectrophotometer

Buy Shimadzu Uv-1800 Uv/Visible Scanning Spectrophotometer Items Including:
  • Constant Temperature 2-Cell Holder for Shimadzu UV-1800 UV/Visible Sca
  • Constant Temperature 4-Cell Holder for Shimadzu UV-1800 UV/Visible Sca
  • Peristaltic Sipper Pump for Shimadzu UV-1800 UV/Visible Scanning Spect.

Graduated Beakers with Handles, Pp

Buy Graduated Beakers with Handles, Pp Items Including:
  • Polypropylene graduated beaker with handle, 1000 mL (V442941)
  • Polypropylene graduated beaker with handle, 2000 mL (V443941)
  • Polypropylene graduated beaker with handle, 3000 mL (V444941)
  • Polypropylene graduated beaker with handle, 5000 mL (V445941).



Allsafe Smc Nemesis Safety Glasses to Graduated Beakers with Handle

>