Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 70
Nilfisk Vacuum Bags 82367805
to
Advantec Grade Gf75 Glass Fiber Filters
Image
Product Detail
More Detail
Nilfisk Vacuum Bags 82367805
Buy Nilfisk Vacuum Bags 82367805 Items Including:
Replacement vacuum bags, pack of 5
Replacement Dust Bags for Nilfisk Backpack HEPA Vacuum, 5/pack.
More Details
Colder Miniature Quick-Disconnect Fittings, Complete Couplings
Buy Colder Miniature Quick-Disconnect Fittings, Complete Couplings Items Including:
Miniature Quick-Disconnect Coupling; Acetal; Straight-through, 1/16" I (SMC01)
Miniature Quick-Disconnect Coupling; PP; Straight-through, 1/16" ID, P (SMC0112)
Miniature Quick-Disconnect Coupling; Acetal; Straight-through, 3 mm ID (SMCM3).
More Details
High-Flow Nylon Monofilament Filter Bags
Buy High-Flow Nylon Monofilament Filter Bags Items Including:
Size 1 high-flow nylon monofilament filter bag; 300°F (149°C) (255050-03)
Size 1 high-flow nylon monofilament filter bag; 300°F (149°C) (255051-03)
Size 1 high-flow nylon monofilament filter bag; 300°F (149°C) (255052-03).
More Details
Nylon Mesh Filter Screen Seattle
Buy Nylon Mesh Filter Screen Seattle Items Including:
Millipore Steriflip Vacuum-Driven Filter Device with Nylon mesh membra
Spectra/Mesh Screen Sheets, 30 X 30 cm Nylon, 100 µm, 3/pk
Spectra/Mesh Screen Sheets, 30 X 30 cm Nylon, 70 µm, 3/pk
Spectra/Mesh Screen Sheets, 30 X 30 cm Nylon, 60 µm, 3/pk.
More Details
Nylon Mesh Filter Screening
Buy Nylon Mesh Filter Screening Items Including:
Spectra/Mesh Screen Sheets, 30 X 30 cm Nylon, 100 µm, 3/pk
Spectra/Mesh Screen Sheets, 30 X 30 cm Nylon, 70 µm, 3/pk
Spectra/Mesh Screen Sheets, 30 X 30 cm Nylon, 60 µm, 3/pk
Spectra/Mesh Screen Discs, 90 mm Nylon, 90mm dia; pore size, 100µ;.
More Details
Digital Micrometers
Buy Digital Micrometers Items Including:
Digital Micrometer (54-815-001-2)
Mechanical Digital Micrometer (52-224-001)
RS-232 cable for micrometer (54-115-779)
Micrometer with digital counter, 0 to 1" range (52-222-001-1)
54-860-001:0-1in/25MM Fluid Resist
More Details
Autoclavable Tubing
Buy Autoclavable Tubing Items Including:
Tubing sets, Autoclavable Pharmed BPT with 2 welded stops for use in 3 (SC0730)
Tubing sets, Autoclavable Pharmed BPT with 2 welded stops for use in 3 (SC0736)
Tubing sets, PharMed BPT with 2 welded stops, 400 mm length, 0.25 mm I (SC0740).
More Details
Crane Scales
Buy Crane Scales Items Including:
Intercomp Lightweight Crane Scale, 50 lb/25 kg
Intercomp Lightweight Crane Scale, 100 lb/50 kg
Fairbanks Hanging Scale, 25 lb x 2 oz (94692)
Fairbanks Hanging Scale, 60 lb x 4 oz (94768).
More Details
800 Mesh Filter Screen
Buy 800 Mesh Filter Screen Items Including:
Spectra/Mesh Screen Sheets, 30 X 30 cm Nylon, 100 µm, 3/pk
Spectra/Mesh Screen Sheets, 30 X 30 cm Nylon, 70 µm, 3/pk
Spectra/Mesh Screen Sheets, 30 X 30 cm Nylon, 60 µm, 3/pk
Spectra/Mesh Screen Discs, 90 mm Nylon, 90mm dia; pore size, 100µ;.
More Details
Glove Box Replacement
Buy Glove Box Replacement Items Including:
Neoprene Gloves for Glove Boxes; size 9 3/4; 30"L (50056-00)
Butyl Gloves; size 8 1/2; 30"L (50055-01)
Butyl Gloves; size 9 3/4; 30"L (50056-01)
Stainless steel support cart for glove box models 34752-20 and -22
Me (800-CART-ABD).
More Details
Glove Box Replacement Gloves
Buy Glove Box Replacement Gloves Items Including:
Neoprene Gloves for Glove Boxes; size 8 1/2; 30"L (5005500)
Butyl Gloves; size 8 1/2; 30"L (50055-01)
Butyl Gloves; size 9 3/4; 30"L (50056-01)
Hypalon Gloves; size 8 1/2; 30"L
Replacement Inlet Or Outlet HEPA Filter (5225105).
More Details
Duran Keck Tubing Quick Disconnect Adapters
Buy Duran Keck Tubing Quick Disconnect Adapters Items Including:
Keck Tubing Adapter, 8 mm to 10 mm ID, 10/pk
Keck Tubing Adapter, 3 mm to 5 mm ID, 10/pk.
More Details
Lightnin Mixer
Buy Lightnin Mixer Items Including:
Lightnin LabMaster Analog Remote Control Mixer with Tube Suspension Mo
Lightnin LabMaster Analog Remote Control Mixer with Universal Clamp Mo
Lightnin® 316 SS high-shear radial flow impeller (Lightnin® R- (YO-04560-30).
More Details
Hepa Vacuum Bags
Buy Hepa Vacuum Bags Items Including:
Replacement Dust Bags for Nilfisk Backpack HEPA Vacuum, 5/pack.
More Details
Spiral Tubing
Buy Spiral Tubing Items Including:
Clear Spiral Wrap,3/16" to 2" (YO-96610-10).
More Details
Ws Tyler Sieve
Buy Ws Tyler Sieve Items Including:
W.S
Tyler Test Stand for Ro-Tap Shakers, 20
W.S
Tyler Wet Test Kit for 8" Diameter Ro-tap Shakers Only
W.S
More Details
Buchi Rotary Evaporators and Accessories
Buy Buchi Rotary Evaporators and Accessories Items Including:
Rotary Evaporator, Diagonal Condenser, Advanced Controller, 220V (YO-23012-01)
Rotary Evaporator, Safety-Coated Vertical Reflux Condenser, Advanced C (YO-23012-36)
Rotary Evaporator, Safety-coated Vertical Reflux Condenser, Profession (YO-23012-37).
More Details
Tube X Compression Adapter
Buy Tube X Compression Adapter Items Including:
Threaded to compression adapter, polypropylene, 5/16" tube OD x 1/4" N (YO-06385-30)
Threaded to compression adapter, polypropylene, 3/8" tube OD x 1/4" NP (YO-06385-40).
More Details
Drum Heaters
Buy Drum Heaters Items Including:
Fifty-five gallon drum heater, 230 VAC (309-55GAL)
9 1/2" wide flexible drum heater, 5 gallon, 1000 watt, 120 VAC, 70-190 (YO-09203-00)
9 1/2" wide flexible drum heater, 15 gallon, 1000 watt, 120 VAC, 70-19 (YO-09203-10).
More Details
Nabertherm Muffle Furnaces
Buy Nabertherm Muffle Furnaces Items Including:
MUFFLE FURNACE, 1460 CU.IN., 100-1100°C, 208 / 240 VAC (LT 24/11/P330)
MUFFLE FURNACE, 2440 CU.IN., 100-1100°C, 208 / 240 VAC (LT 40/11/P330)
MUFFLE FURNACE, 1460 CU.IN., 100-1200°C, 208 / 240 VAC (LT 24/12/P330).
More Details
Parker Hannifin Pfa Straight Compression Unions
Buy Parker Hannifin Pfa Straight Compression Unions Items Including:
Straight compression union, PFA, 1/8" tubing OD (YO-31320-21)
Straight compression union, PFA, 1/4" tubing OD (YO-31320-22)
Straight compression union, PFA, 3/8" tubing OD (YO-31320-23).
More Details
Urine and Saliva Ph Test Paper
Buy Urine and Saliva Ph Test Paper Items Including:
Universal pH Test Paper; pH Range 1 to 11; 10 Booklets of 20 Strips
Universal pH Test Paper; pH Range 1 to 11; 9 mm x 6 m Roll
Litmus Blue Test Paper, 10 Booklets of 20 Strips
Litmus Red Test Paper, 10 Booklets of 20 Strips
Jumbo pH paper, 1 - 12.
More Details
Asti all-Pfa Double-Bellows Pumps
Buy Asti all-Pfa Double-Bellows Pumps Items Including:
Asti All (PFA-coated) Double-Bellows Pumps 10 LPM
Asti All (PFA-coated) Double-Bellows Pumps 30 LPM, 5 bar, 3/4" flare c.
More Details
Luxo Illuminated Magnifiers
Buy Luxo Illuminated Magnifiers Items Including:
10-Diopter Swing-away Auxiliary Lens.
More Details
Best Latex Gloves
Buy Best Latex Gloves Items Including:
Best Derma Thin Latex Gloves, small
Best HD™ Liquid-Proof Latex Gloves, Rolled Cuff, large, 15"L, 12 pa
Best HD™ Liquid-Proof Latex Gloves, Rolled Cuff, extra-large, 15"L,
Best N-DEX PLUS Non-latex Industrial Gloves; size, small; 50/pack.
More Details
Aalborg Valves
Buy Aalborg Valves Items Including:
Aalborg Stepping Motor Proportioning Valve (aluminum), 200 sL/min (air
Aalborg Stepping Motor Proportioning Valve (stainless steel), 200 sL/m
Aalborg Stepping Motor Proportioning Valve (aluminum), 500 sL/min (air.
More Details
Thermolyne Muffle Furnaces
Buy Thermolyne Muffle Furnaces Items Including:
Thermo Scientific Thermolyne Benchtop Muffle Furnace with 120 cu in ch (F47910-33)
Thermo Scientific Thermolyne Benchtop Muffle Furnace with 350 cu in ch (F48015-60)
Thermo Scientific Thermolyne Benchtop Muffle Furnace with 120 cu in ch (F47925-80).
More Details
Wide-Mouth Bottles to 1000 Ml
Buy Wide-Mouth Bottles to 1000 Ml Items Including:
Thermo Scientific Nalgene low-density polyethylene wide-mouth bottle, (2103-0032)
Thermo Scientific Nalgene low-density polyethylene wide-mouth bottle, (2103-0001)
Thermo Scientific Nalgene Wide-Mouth Economy Bottle, PPCO, 1000 mL, 6/ (YO-06034-71).
More Details
Beckman Ph Meter Psi 310
Buy Beckman Ph Meter Psi 310 Items Including:
Oakton pH 310 meter with single-junction pH/ATC probe
Oakton pH 310 meter kit with NIST-traceable Calibration
pH electrode sealed single-junction for Beckman meters (YO-27507-00)
pH electrode sealed double-junction for Beckman meters (YO-27507-02).
More Details
Advantec Grade Gf75 Glass Fiber Filters
Buy Advantec Grade Gf75 Glass Fiber Filters Items Including:
Advantec Grade GF75 Glass Fiber Filters; 47mm dia; 100/box
Advantec Grade GF75 Glass Fiber Filters; 90mm dia; 100/box
Advantec Grade GF75 Glass Fiber Filters; 125mm dia; 100/box
Advantec Grade GF75 Glass Fiber Filters; 142mm dia; 100/box.
More Details
70618
results
<<<Previous
66
67
68
69
70
71
72
73
74
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
20
30
40
50
55
56
57
58
59
60
61
62
63
64
65
66
67
68
69
70
71
72
73
74
75
76
77
78
79
80
81
82
83
84
90
100
110
120
130
140
150
160
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Nilfisk Vacuum Bags 82367805 to Advantec Grade Gf75 Glass Fiber Filters
>
31125329-s-toc_2 31125329stoc2