Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 700

53mm Bottle Top Dispenser   to   Ise Standard Solutions

Image
  Product Detail
More Detail

53mm Bottle Top Dispenser

Buy 53mm Bottle Top Dispenser Items Including:
  • Cole-Parmer Bottletop Dispenser .25-2.5 mL
  • Cole-Parmer Bottletop Dispenser .25-2.5 mL with 100 mL Glass Bottle
  • Cole-Parmer Bottletop Dispenser 2.5-30 mL with 500 mL Glass Bottle
  • Cole-Parmer Bottletop Dispenser 5-60 mL with 1000 mL Glass Bottle.

Laboratory Equipment Syringes

Buy Laboratory Equipment Syringes Items Including:
  • Cole-Parmer HPLC Syringe Filters, Non Sterile, Nylon, 17mm, 0.45um 100
  • Cole-Parmer HPLC Syringe Filters, Non Sterile, Nylon, 0.45 um, 30 mm d (YO-32816-04)
  • Cole-Parmer HPLC Syringe Filters, Non Sterile, Nylon, 0.45 um, 30 mm d (YO-32816-06).

Rotary Vane Compressor Pumps

Buy Rotary Vane Compressor Pumps Items Including:
  • Vacuubrandreg Rotary Vane Vacuum Pump, 1.6 cfm, 1.5 X 10-3/sup (698123)
  • Vacuubrandreg Rotary Vane Vacuum Pump, 1.6 cfm, 1.5 X 10-3/sup (698120)
  • Vacuubrandreg Rotary Vane Vacuum Pump, 4.0 cfm, 1.5 X 10-3/sup (698133).

Mixers Impeller

Buy Mixers Impeller Items Including:
  • Compact straight blade impeller; 1-1/4" dia
  • Compact square blade impeller; 1-1/4" dia
  • Square blade impeller; 4" dia
  • Cross blade impeller; 2" dia
  • Collapsible blade impeller; 3-1/2" dia

E. Coli 0157 Test Kits

Buy E. Coli 0157 Test Kits Items Including:
  • CT-supplement, 16 vials (YO-36152-28)
  • mEC selective broth with Novobiocin, 500 g (YO-36152-22)
  • mTSB selective broth with Novobiocin, 500g (YO-36152-24).

Aluminum Filter Bag Housing

Buy Aluminum Filter Bag Housing Items Including:
  • General-purpose 304 stainless steel filter bag housing, 1-1/4" NPT(F) (156120)
  • General-purpose 304 stainless steel filter bag housing, 2" NPT(F) (156121)
  • General-purpose 304 stainless steel filter bag housing, 2" NPT(F) (156122).

Polyester Cleanroom Swab

Buy Polyester Cleanroom Swab Items Including:
  • Cleanroom swab; fabric with knit polyester tip and sealed polypropylen (TX759B)
  • Cleanroom swab; fabric with knit polyester tip and polypropylene handl (TX743B)
  • Cleanroom swab; fabric with knit polyester tip and polypropylene handl (TX714A).

Sulfuric Acid Filtration what Filter Paper

Buy Sulfuric Acid Filtration what Filter Paper Items Including:
  • Whatman Glass Microfiber Filter Papers Water-Pollution test papers; 4
  • (YO-06648-70)
  • Whatman Glass Microfiber Filter Papers Water-Pollution test papers; 4
  • (YO-06648-71).

Distillation Apparatus Stainless Steel

Buy Distillation Apparatus Stainless Steel Items Including:
  • Jiffy-Jack Lab Jack, Heavy-Duty Apparatus Positioners, 20lb (9.0kg) li
  • Jiffy-Jack Lab Jack, Heavy-Duty Apparatus Positioners, 30lb (13.5kg) l
  • Frame Rods, 9-13/16" (250mm), 2/pack (53-2522)
  • Frame Rods, 29-1/2" (750 mm) (YO-08024-24).

Meterman Lcr - Cr Meters

Buy Meterman Lcr - Cr Meters Items Including:
  • Capacitance and Resistance Meter from Amprobe
  • LCR Meter with transistor test (LCR55A).

Gates Terminator Multi-Purpose Hose

Buy Gates Terminator Multi-Purpose Hose Items Including:
  • Gates Terminator Multi-Purpose Hose; ¼" x 0.57", 100 ft/pk
  • Gates Terminator Multi-Purpose Hose; ½" x 0.89", 100 ft/pk
  • Gates Terminator Multi-Purpose Hose; 1-½" x 2.08", 25 ft/pk.

Temptale 4 Temperature Control

Buy Temptale 4 Temperature Control Items Including:
  • Love Temperature Control, 1/4 DIN, Temperature Input, Voltage Pulse Ou (4C-2)
  • Temperature Controller, TC, 1/4-DIN, Relay Output, 2 alarms, RS485 (DC1040CT-102-002-E)
  • Temperature Controller, RTD, 1/4-DIN, Relay Output, 1 alarms, (DC1044CR-101-000-E).

5x Magnifier

Buy 5x Magnifier Items Including:
  • Bausch & Lomb Folding Pocket Magnifier, triple lens, 5x to 20x magnifi
  • Clip-On Magnifier, Large frame, 5x magnification
  • Clip-On Monocular, Single Lens, 5x (JF5X).

Nunc General Accessories

Buy Nunc General Accessories Items Including:
  • Nonsterile lids for Thermo Scientific Nunc™ 96-well plates 01929-32 (YO-01929-10)
  • Sterile lids for Thermo Scientific Nunc™ 96-well plates 01929-32, - (YO-01929-11)
  • 96-Well Caps for Thermo Scientific Nunc™ 96-Well Microplates, Steri (YO-01929-54).

Bronze Gear Pump Head

Buy Bronze Gear Pump Head Items Including:
  • Bronze Gear Pump Head, Close-Coupled Design, 1.0 GPM at 0 PSI when use
  • Bronze Gear Pump Head, Close-Coupled Design, 1GPM at 0 PSI when used w
  • Bronze Gear Pump Head Motor Adapter for NEMA 56C
  • Bronze Gear Pump Head Motor Adapter for IEC motors.

Cole-Parmer Wide-Mouth Pet Jars

Buy Cole-Parmer Wide-Mouth Pet Jars Items Including:
  • PET wide-mouth jar, 180 mL (107006)
  • PET wide-mouth jar, 240 mL (107008)
  • PET wide-mouth jar, 350 mL (107012)
  • PET wide-mouth jar, 480 mL (107016A)
  • PET wide-mouth jar, 960 mL (107032E)

Visual Laboratory Storage Cabinets

Buy Visual Laboratory Storage Cabinets Items Including:
  • Standard cabinet (CV1252TAN).

4khj3 Bronze Rotary Gear Pump Head

Buy 4khj3 Bronze Rotary Gear Pump Head Items Including:
  • Bronze Gear Pump Head, Close-Coupled Design, 1.0 GPM at 0 PSI when use
  • Bronze Gear Pump Head, Close-Coupled Design, 1GPM at 0 PSI when used w
  • Bronze Gear Pump Head Motor Adapter for NEMA 56C
  • Bronze Gear Pump Head Motor Adapter for IEC motors.

Kestrel Weather Meters

Buy Kestrel Weather Meters Items Including:
  • Pocket Weather Monitor (KESTREL-4000)
  • Kestrel 08304USB USB Interface Cradle for 4000 Series Handheld Meters
  • Kestrel 1000 Pocket Wind Meter; Measures Wind Speed Maxwind Gust And
  • Kestrel 2000 Pocket Wind Meter; Measures Wind Speed Temperature Wind.

Plug Valves

Buy Plug Valves Items Including:
  • Plug Valve with Pipe Thread Connections, 1/4" NPT(F) (D1071794)
  • Valve Plug PFA 1/8"OD (204-22)
  • Valve Plug PFA 1/4"OD (204-19)
  • Polypropylene Dummy Plug
  • Float Switch with Piggyback Plug.

Ultrasonication Probes

Buy Ultrasonication Probes Items Including:
  • Threaded probes with replaceable 1 (YO-04710-35)
  • Temperature probe, stainless steel (YO-04711-50)
  • Ultrasonic Cell Disruptor Replacement Probes with Threaded Tips, 1/2"
  • Ultrasonic Cell Disruptor Replacement Probes with Threaded Tips, 3/4".

Thermo Conductivity Cells

Buy Thermo Conductivity Cells Items Including:
  • Thermo Scientific AquaPro Conductivity Sensor
  • Thermo Scientific Orion 4-Cell Conductivity Probe, K = 0.475, 5-ft cab
  • Thermo Scientific Orion 2-Cell Conductivity Electrode, K=0.1, 5-ft cab.

304 Stainless Steel Sanitary Clamp

Buy 304 Stainless Steel Sanitary Clamp Items Including:
  • Cole-Parmer, Threaded Sanitary Clamp Adapter, 304L SS, 1" Clamp X 3/4"
  • Cole-Parmer, Threaded Sanitary Clamp Adapter, 304L SS, 1.5" Clamp X 3/
  • Cole-Parmer, Threaded Sanitary Clamp Adapter, 304L SS, 1.5" Clamp X 1
  • (YO-30800-59).

Ysi Dissolved Oxygen, Bod, and Cod

Buy Ysi Dissolved Oxygen, Bod, and Cod Items Including:
  • YSI Model 550A Dissolved Oxygen Meter, 12-ft (3.7-m) probe cables
  • YSI Model 550A Dissolved Oxygen Meter, 25-ft (7.6-m) probe cables
  • YSI DO 200 Dissolved Oxygen Meter Only
  • YSI Pro20 Dissolved Oxygen Meter
  • YSI ProODO™ Optical Dissolved Oxygen Meter.

Ball Valve Sanitary Fitting

Buy Ball Valve Sanitary Fitting Items Including:
  • Sanitary Ball Valve, 1-1/2" TRI-CLAMP
  • Sanitary Butterfly Valve, 1" TRI-CLAMP
  • Sanitary Butterfly Valve, 1-1/2" TRI-CLAMP
  • Sanitary Butterfly Valve, 4" TRI-CLAMP
  • Sanitary Butterfly Valve, 3" TRI-CLAMP.

Dwyer Air Velocity and Pressure Meters

Buy Dwyer Air Velocity and Pressure Meters Items Including:
  • 4005LKIT:Air Vlcty 0/5 Inwc Air Velocity M Kit 400/9000 Velocity Fpm W
  • 40023KIT:Airvlcty 0/23 Inwc Air Velocity M Kit 400/19 200 Velocity Fpm.

Thermo Scientific Nanodrop

Buy Thermo Scientific Nanodrop Items Including:
  • Thermo Scientific NanoDrop ND-8000 8-position Spectrophotometer
  • Thermo Scientific NanoDrop™ ND-2000c Spectrophotometer with Cuvett
  • Thermo Scientific NanoDrop™ ND-2000 1-position Spectrophotometer wi
  • Calibration fluid for Thermo Scientific NanoDrop.

Dow Corning Tubing, and Hose

Buy Dow Corning Tubing, and Hose Items Including:
  • Dow Corning Pharma-50 Tubing, 3/16" x 5/16", 50-ft pk.

Cole-Parmer 12-Mm Orp Electrodes

Buy Cole-Parmer 12-Mm Orp Electrodes Items Including:
  • Cole-Parmer Single-Junction ORP Electrode, Platinum Sensor, 10-ft Cabl
  • Cole-Parmer 12-mm Diameter, Double-Junction Atimony pH Electrode (YO-05994-06)
  • Cole-Parmer 12-mm Diameter, Single-Junction Atimony pH Electrode (YO-05994-24).

Ise Standard Solutions

Buy Ise Standard Solutions Items Including:
  • ISE Standard solution, chloride, 0.1 M (YO-27503-08)
  • ISE Standard solution, fluoride, 0.1 M (YO-27503-12)
  • Sodium (Na+) ISE Double junction solution kits (YO-27502-93)
  • Surfactant (X+/X-) ISE Double junction solution (YO-27502-95).



53mm Bottle Top Dispenser to Ise Standard Solutions

>