Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Product Index Page 786
Magnetic Stir Bar Pull
to
Refrigerated Centrifuge 3000 Rpm
Image
Product Detail
More Detail
Magnetic Stir Bar Pull
Buy Magnetic Stir Bar Pull Items Including:
Spinbar Magnetic Stir Bar Rack
Glass Encased magnetic Stir Bars, 1/2"l X 3/8"d (F37101-0012)
Glass Encased magnetic Stir Bars, 1"l X 3/8"d (F37101-0001)
Glass Encased magnetic Stir Bars, 1 1/2"l X 3/8"d (F37101-0112).
More Details
Magnetic Stir Bar Sets
Buy Magnetic Stir Bar Sets Items Including:
Spinbar Magnetic Stir Bar Rack
Glass Encased magnetic Stir Bars, 1/2"l X 3/8"d (F37101-0012)
Glass Encased magnetic Stir Bars, 1"l X 3/8"d (F37101-0001)
Glass Encased magnetic Stir Bars, 1 1/2"l X 3/8"d (F37101-0112).
More Details
Magnetic Stir Bar Substitute
Buy Magnetic Stir Bar Substitute Items Including:
Spinbar Magnetic Stir Bar Rack
Glass Encased magnetic Stir Bars, 1/2"l X 3/8"d (F37101-0012)
Glass Encased magnetic Stir Bars, 1"l X 3/8"d (F37101-0001)
Glass Encased magnetic Stir Bars, 1 1/2"l X 3/8"d (F37101-0112).
More Details
Magnetic Stir Bar Weight
Buy Magnetic Stir Bar Weight Items Including:
Spinbar Magnetic Stir Bar Rack
Glass Encased magnetic Stir Bars, 1/2"l X 3/8"d (F37101-0012)
Glass Encased magnetic Stir Bars, 1"l X 3/8"d (F37101-0001)
Glass Encased magnetic Stir Bars, 1 1/2"l X 3/8"d (F37101-0112).
More Details
Magnetic Stir Bars Assortment
Buy Magnetic Stir Bars Assortment Items Including:
Spinbar Magnetic Stir Bar Rack
Glass Encased magnetic Stir Bars, 1/2"l X 3/8"d (F37101-0012)
Glass Encased magnetic Stir Bars, 1"l X 3/8"d (F37101-0001)
Glass Encased magnetic Stir Bars, 1 1/2"l X 3/8"d (F37101-0112).
More Details
Magnetic Stir Bars Patenet
Buy Magnetic Stir Bars Patenet Items Including:
Spinbar Magnetic Stir Bar Rack
Glass Encased magnetic Stir Bars, 1/2"l X 3/8"d (F37101-0012)
Glass Encased magnetic Stir Bars, 1"l X 3/8"d (F37101-0001)
Glass Encased magnetic Stir Bars, 1 1/2"l X 3/8"d (F37101-0112).
More Details
Magnetic Stir Bars Review
Buy Magnetic Stir Bars Review Items Including:
Spinbar Magnetic Stir Bar Rack
Glass Encased magnetic Stir Bars, 1/2"l X 3/8"d (F37101-0012)
Glass Encased magnetic Stir Bars, 1"l X 3/8"d (F37101-0001)
Glass Encased magnetic Stir Bars, 1 1/2"l X 3/8"d (F37101-0112).
More Details
Magnetic Stirring Bars by Fisher Sci
Buy Magnetic Stirring Bars by Fisher Sci Items Including:
Spinbar Magnetic Stir Bar Rack
Glass Encased magnetic Stir Bars, 1/2"l X 3/8"d (F37101-0012)
Glass Encased magnetic Stir Bars, 1"l X 3/8"d (F37101-0001)
Glass Encased magnetic Stir Bars, 1 1/2"l X 3/8"d (F37101-0112).
More Details
Microtiter Magnetic Stir Bar
Buy Microtiter Magnetic Stir Bar Items Including:
Spinbar Magnetic Stir Bar Rack
Glass Encased magnetic Stir Bars, 1/2"l X 3/8"d (F37101-0012)
Glass Encased magnetic Stir Bars, 1"l X 3/8"d (F37101-0001)
Glass Encased magnetic Stir Bars, 1 1/2"l X 3/8"d (F37101-0112).
More Details
Non Magnetic Stir Bar
Buy Non Magnetic Stir Bar Items Including:
Spinbar Magnetic Stir Bar Rack
Glass Encased magnetic Stir Bars, 1/2"l X 3/8"d (F37101-0012)
Glass Encased magnetic Stir Bars, 1"l X 3/8"d (F37101-0001)
Glass Encased magnetic Stir Bars, 1 1/2"l X 3/8"d (F37101-0112).
More Details
Octogonal Magnetic Stir Bar
Buy Octogonal Magnetic Stir Bar Items Including:
Spinbar Magnetic Stir Bar Rack
Glass Encased magnetic Stir Bars, 1/2"l X 3/8"d (F37101-0012)
Glass Encased magnetic Stir Bars, 1"l X 3/8"d (F37101-0001)
Glass Encased magnetic Stir Bars, 1 1/2"l X 3/8"d (F37101-0112).
More Details
Smallest Magnetic Stirring Bars
Buy Smallest Magnetic Stirring Bars Items Including:
Spinbar Magnetic Stir Bar Rack
Glass Encased magnetic Stir Bars, 1/2"l X 3/8"d (F37101-0012)
Glass Encased magnetic Stir Bars, 1"l X 3/8"d (F37101-0001)
Glass Encased magnetic Stir Bars, 1 1/2"l X 3/8"d (F37101-0112).
More Details
Sterile Magnetic Stir Bar
Buy Sterile Magnetic Stir Bar Items Including:
Spinbar Magnetic Stir Bar Rack
Glass Encased magnetic Stir Bars, 1/2"l X 3/8"d (F37101-0012)
Glass Encased magnetic Stir Bars, 1"l X 3/8"d (F37101-0001)
Glass Encased magnetic Stir Bars, 1 1/2"l X 3/8"d (F37101-0112).
More Details
Tiny Magnetic Stir Bars
Buy Tiny Magnetic Stir Bars Items Including:
Spinbar Magnetic Stir Bar Rack
Glass Encased magnetic Stir Bars, 1/2"l X 3/8"d (F37101-0012)
Glass Encased magnetic Stir Bars, 1"l X 3/8"d (F37101-0001)
Glass Encased magnetic Stir Bars, 1 1/2"l X 3/8"d (F37101-0112).
More Details
Vwr Magnetic Stir Bars
Buy Vwr Magnetic Stir Bars Items Including:
Spinbar Magnetic Stir Bar Rack
Glass Encased magnetic Stir Bars, 1/2"l X 3/8"d (F37101-0012)
Glass Encased magnetic Stir Bars, 1"l X 3/8"d (F37101-0001)
Glass Encased magnetic Stir Bars, 1 1/2"l X 3/8"d (F37101-0112).
More Details
Ph Probe Testing Kit
Buy Ph Probe Testing Kit Items Including:
Aqua Comparator Test Kit, Nitrate, 10-100 mg/L (L147400)
CHEMets Colorimetric Dissolved Oxygen Test Kit, 0 to 40 ppb (K-7540)
CHEMets Colorimetric Dissolved Oxygen Test Kit, 0 to 100 ppb (K-7599)
Microbial Test Kits, Aerobic Bateria Test Kit (110).
More Details
Ph Sticks Liquid Test Kits
Buy Ph Sticks Liquid Test Kits Items Including:
Aqua Comparator Test Kit, Nitrate, 10-100 mg/L (L147400)
CHEMets Colorimetric Dissolved Oxygen Test Kit, 0 to 40 ppb (K-7540)
CHEMets Colorimetric Dissolved Oxygen Test Kit, 0 to 100 ppb (K-7599)
Microbial Test Kits, Aerobic Bateria Test Kit (110).
More Details
Electrophoresis Horizontal Systems
Buy Electrophoresis Horizontal Systems Items Including:
Additional Gel Tray for Cole-Parmer Horizontal Mini-Gel Systems, for M (YO-28559-70)
Additional Gel Tray for Cole-Parmer Horizontal Mini-Gel Systems, for M (YO-28559-72)
Gel Tray for Cole-Parmer Horizontal Mid-Size Gel System Model 28560-00.
More Details
Goggles Safety Uvex
Buy Goggles Safety Uvex Items Including:
Uvex STEALTH Safety Goggles, Clear Lens
Uvex Strategy Safety Goggles with Fabric Band, Gray Body with Direct V
Uvex Strategy Safety Goggles with Neoprene Band, Gray Body with Indire
Flex Seal Safety Goggle, gray lens (S3410X)
Laser Goggles, HE-NE.
More Details
Purefit Masterflex Tubing and Pump Accessories
Buy Purefit Masterflex Tubing and Pump Accessories Items Including:
PureFit™ TC Tubing Clamp 3/8" Polypropylene, 10/pk (YO-06822-01)
PureFit™ TC Tubing Clamp 3/8" PVDF, 10/pk (YO-06822-03)
PureFit™ TC Tubing Clamp 3/4" Polypropylene, 10/pk (YO-06822-11).
More Details
Abb Commander 100
Buy Abb Commander 100 Items Including:
Chart Paper for ABB Commander 250-mm Strip Chart Recorders, 100 divisi.
More Details
Easun Pneumatic Push Fittings
Buy Easun Pneumatic Push Fittings Items Including:
SMC pneumatic elbow union, 1/4" (MS-5P)
SMC pneumatic Y splitter, 1/4" (KJU07-00)
SMC pneumatic straight union, 1/4" (KQ2H07-00).
More Details
General Tool Stainless Steel Digital Calipers
Buy General Tool Stainless Steel Digital Calipers Items Including:
Digital Stainless Steel Caliper, 0 to 6" with Fractions," or Milimeter (147)
Digital Stainless Steel Caliper, 0 to 8" with Fractions," or Milimeter (1478).
More Details
Hvac/R Digital and Analog Manometers
Buy Hvac/R Digital and Analog Manometers Items Including:
Datalogging Manometer, 0 To 2 PSI
Datalogging Manometer, 0 To 5 PSI
Datalogging Manometer, 0 To 15 PSI
Ac Adapter For Datalogging Manometers 68603-01, -03, -05.
More Details
Pneumatic Push Fittings Rochester Ny
Buy Pneumatic Push Fittings Rochester Ny Items Including:
SMC pneumatic elbow union, 1/4" (MS-5P)
SMC pneumatic Y splitter, 1/4" (KJU07-00)
SMC pneumatic straight union, 1/4" (KQ2H07-00).
More Details
Pneumatic Push in Fittings
Buy Pneumatic Push in Fittings Items Including:
SMC pneumatic elbow union, 1/4" (MS-5P)
SMC pneumatic Y splitter, 1/4" (KJU07-00)
SMC pneumatic straight union, 1/4" (KQ2H07-00).
More Details
Sperian Willson Safety Glasses
Buy Sperian Willson Safety Glasses Items Including:
SPERIAN MILLENNIA™ Protective Glasses, Clear Lens, Black Frame.
More Details
4 Foot Glass Cylinders
Buy 4 Foot Glass Cylinders Items Including:
Pyrexplus Brand 63024 cylinder; 100 mL, case of 4
Pyrexplus Brand 63024 cylinder; 250 mL, case of 4
Pyrexplus Brand 63024 cylinder; 500 mL, case of 4
Pyrexplus Brand 63024 cylinder; 1000 mL, case of 4.
More Details
Threaded Fitting Tube Connector
Buy Threaded Fitting Tube Connector Items Including:
Pipe fittings, Bulkhead connectors, PVC, 1/4" NPT (F) x 27/32" OD1-3/8 (YO-06445-10)
Bulkhead Fitting, PTFE, 1/4" NPT(F) Port X 1-11/16" OD (F-TA02)
Bulkhead Fitting, PTFE, 3/8" NPT(F) Port X 1-13/16" OD (F-TA03).
More Details
Refrigerated Centrifuge 3000 Rpm
Buy Refrigerated Centrifuge 3000 Rpm Items Including:
Thermo Scientific Hereaus X1R Refrigerated Centrifuge, to 15,200 rpm, (YO-17709-06)
Thermo Scientific Hereaus X1R Refrigerated Centrifuge, to 15,200 rpm, (YO-17709-08)
Eppendorf multi-purpose centrifuge; 4 x 250 mL capacity, 115 VAC (YO-02570-10).
More Details
70998
results
<<<Previous
782
783
784
785
786
787
788
789
790
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
100
200
300
400
500
680
690
700
710
720
730
740
750
760
771
772
773
774
775
776
777
778
779
780
781
782
783
784
785
786
787
788
789
790
791
792
793
794
795
796
797
798
799
800
810
820
830
840
850
860
870
880
900
1,000
1,100
1,200
1,300
1,400
1,500
1,600
1,700
2,000
2,367
Magnetic Stir Bar Pull to Refrigerated Centrifuge 3000 Rpm
>
31125329-s-toc_2 31125329stoc2