Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 8
Dissolved Oxygen Meter
to
Gilmont Micrometer Syringe
Image
Product Detail
More Detail
Dissolved Oxygen Meter
Buy Dissolved Oxygen Meter Items Including:
YSI splashproof dissolved oxygen meter with probe, 25-ft cable, model
YSI Model 550A Dissolved Oxygen Meter, 12-ft (3.7-m) probe cables
YSI Model 5000 Benchtop Dissolved Oxygen Meter, 115 VAC
YSI Model 5100 Benchtop Dissolved Oxygen Meter, 115 VAC.
More Details
Cole-Parmer Barbed Cross Connectors
Buy Cole-Parmer Barbed Cross Connectors Items Including:
Barbed Fittings; Cross Connector, 316 SS, 1/16" Tubing ID (YO-31208-51)
Barbed Fittings; Cross Connector, 316 SS, 3/32" Tubing ID (YO-31208-52)
Barbed Fittings; Cross Connector, 316 SS, 3/16" Tubing ID (YO-31208-54).
More Details
Equipment Calibration Services
Buy Equipment Calibration Services Items Including:
Innocal NIST Calibration for Refrigerators & Freezers
Innocal NIST Calibration for Incubators.
More Details
Sample Containers
Buy Sample Containers Items Including:
Hinged-Lid Sample Containers, PP, 1/4 oz, 2500/pk
Cole-Parmer Lids, PP, for Sample Containers 0610012 & 0610014, 500/cs
Wide-Mouth Sample Containers, PP, 120 mL
Wide-Mouth Sample Containers, PP, 240 mL
Wide-Mouth Sample Containers, PP, 480 mL.
More Details
Vibration Damping Mounts
Buy Vibration Damping Mounts Items Including:
Vibrasorb Damping Mount
Vibrasorb Mount, Large.
More Details
Cole-Parmer Fluoride-Resistant Ph Electrodes
Buy Cole-Parmer Fluoride-Resistant Ph Electrodes Items Including:
Cole-Parmer HF Resistant Submersible pH Electrode, CPVC (YO-27108-00)
Cole-Parmer HF Resistant Submersible pH Electrode, CPVC, 100Ω RT (YO-27108-02)
Cole-Parmer HF Resistant Submersible pH Electrode, CPVC, 1 kΩ RT (YO-27108-04).
More Details
Plastic Check Valves
Buy Plastic Check Valves Items Including:
Hayward True Union Ball Check Valve, 1/2" socket and NPT(F) connectors
Hayward True Union Ball Check Valve, 3/4" socket and NPT(F) connectors
Hayward True Union Ball Check Valve, 1-1/2" socket and NPT(F) connecto.
More Details
Cole-Parmer Luer Fittings and Adapters
Buy Cole-Parmer Luer Fittings and Adapters Items Including:
Luer Fitting Kit, 477 pieces (YO-45511-00)
Cole-Parmer Animal Free Female Luer Fittings, 1/8", PP, 25/pk (YO-30800-02)
Cole-Parmer Animal Free Female Luer Fittings, 3/16", PP, 25/pk
Cole-Parmer Animal Free Female Luer Fittings, PP, 25/pk.
More Details
Cole-Parmer Direct Coupled Rotary Vane Pumps
Buy Cole-Parmer Direct Coupled Rotary Vane Pumps Items Including:
High-pressure, brass-bodied rotary vane pump, 0.75 GPM, 115/230 VAC
High-pressure, brass-bodied rotary vane pump, 2.3 GPM, 115/230 VAC, 5
High-pressure, 303 stainless steel rotary vane pump, 2.3 GPM, 115/230.
More Details
Volumetric Flasks
Buy Volumetric Flasks Items Including:
Thermo Scientific Nalgene Class B polypropylene volumetric flask, 50 m
Thermo Scientific Nalgene Class B polypropylene volumetric flask, 100
PYREX Brand 5640 Volumetric Flask; 10 mL, case of 12
PYREX Brand 5640 Volumetric Flask; 100 mL, case of 12.
More Details
150 Ml Glass Volumetric Flask
Buy 150 Ml Glass Volumetric Flask Items Including:
PYREX Brand 5640 Volumetric Flask; 100 mL, pack of 6
PYREX Brand 5640 Volumetric Flask; 200 mL, pack of 6
PYREX Brand 5640 Volumetric Flask; 1000 mL, pack of 1
PYREX Brand 5640 Volumetric Flask; 2000 mL, pack of 1.
More Details
Thermo Scientific Nalgene Polypropylene Trays
Buy Thermo Scientific Nalgene Polypropylene Trays Items Including:
Thermo Scientific Nalgene polypropylene autoclaving tray, 1 L
Thermo Scientific Nalgene polypropylene autoclaving tray, 2 L
Thermo Scientific Nalgene polypropylene autoclaving tray, 3 L
Thermo Scientific Nalgene polypropylene autoclaving tray, 5 L.
More Details
Asco Scientific Asco Subminiature Solenoid Valves
Buy Asco Scientific Asco Subminiature Solenoid Valves Items Including:
ASCO Subminiature Solenoid Valve 1/8" NPT(F), 3/32" Orfice, 24VDC
ASCO Subminiature Solenoid Valve 1/8" NPT(F), 7/64" Orfice, 120VAC
ASCO Subminiature Solenoid Valve 1/4" NPT(F), 7/64" Orfice, 120VAC.
More Details
Eppendorf Fixed-Volume 2000 Series Pipettors
Buy Eppendorf Fixed-Volume 2000 Series Pipettors Items Including:
Fixed-volume pipettors, 100 µL (22 47 115-5)
Fixed-volume pipettors, 200 µL (22 47 125-2)
Fixed-volume pipettors, 500 µL (22 47 145-7)
Fixed-volume pipettors, 1000 µL (22 47 165-1)
Fixed-volume pipettors, 1500 µL (22 47 170-8).
More Details
John Guest Fitting
Buy John Guest Fitting Items Including:
Quick-disconnect fittings, Standard John Guest valved Fittings, Delri (PLCD14004)
Quick-disconnect fittings, Standard John Guest valved fittings, Delrin (PLCD14006)
Quick-disconnect fittings, Panel-mount John Guest valved fittingDelrin (PLCD11004).
More Details
A&D Weighing Gf Toploading Balances
Buy A&D Weighing Gf Toploading Balances Items Including:
A&D GF Toploading Balance, 410g x 0.001g External Calibration, RS232,
A&D GF Toploading Balance, 610g x 0.001g External Calibration, RS232,
A&D GF Toploading Balance, 810g x 0.001g External Calibration, RS232,.
More Details
Hach Conductivity/Resistivity/Tds Meters and Testers
Buy Hach Conductivity/Resistivity/Tds Meters and Testers Items Including:
Hach® sensION™ 5 Waterproof Conductivity Meter
Hach SensION™+ Rugged Conductivity Meter Kit
Hach SensION™+ Conductivity Meter Kit
Cond
Cell, General.
More Details
Cole-Parmer Refractive Index Standard Solutions
Buy Cole-Parmer Refractive Index Standard Solutions Items Including:
Refractive index standard solution; 1.300 to 1.395, 20 liquids
Refractive index standard solution; 1.300 to 1.390, 10 liquids
Refractive index standard solution; 1.400 to 1.458, 30 liquids.
More Details
Advantec Mixed Cellulose Ester Membrane Filters
Buy Advantec Mixed Cellulose Ester Membrane Filters Items Including:
Advantec Sterile Mixed Cellulose Ester Filter Membranes; 47mm dia; 0.6 (YO-06644-06)
Advantec Sterile Mixed Cellulose Ester Filter Membranes; 47mm dia; 0.4 (YO-06644-08).
More Details
Piston Vacuum Pump
Buy Piston Vacuum Pump Items Including:
Standard Duty Dry Vacuum Piston Pumps, 22 L/min, 115 VAC
Standard Duty Dry Vacuum Piston Pumps, 19 L/min, 230 VAC
Standard Duty Dry Vacuum Piston Pumps, 34 L/min, 115 VAC
Standard Duty Dry Vacuum Piston Pumps, 201 L/min, 115 VAC.
More Details
Magnetic Stir Bars
Buy Magnetic Stir Bars Items Including:
Spinbar Magnetic Stir Bar Rack
Glass Encased magnetic Stir Bars, 1/2"l X 3/8"d (F37101-0012)
Glass Encased magnetic Stir Bars, 1"l X 3/8"d (F37101-0001)
Glass Encased magnetic Stir Bars, 1 1/2"l X 3/8"d (F37101-0112).
More Details
Sanitary Centrifugal Pumps
Buy Sanitary Centrifugal Pumps Items Including:
Sanitary Centrifugal Pump, Heavy-Duty, 50 GPM, 1 HP, White Epoxy Motor
Sanitary Centrifugal Pump, Heavy-Duty, 55 GPM, 1.5 HP, White Epoxy Mot
Sanitary Centrifugal Pump, Heavy-Duty, 110 GPM, 3 HP, White Epoxy Moto.
More Details
Oakton Ph Meter
Buy Oakton Ph Meter Items Including:
Oakton pH 700 Benchtop Meter, double-junction refillable glass pH elec (pH 700) (YO-35419-10)
Oakton pH 700 Benchtop Meter, meter and probe stand
Oakton pH 11 meter only
Oakton pH 11 meter kit
Oakton pH 300 meter with single-junction pH/ATC probe.
More Details
304 Stainless Steel Funnel
Buy 304 Stainless Steel Funnel Items Including:
304 Stainless Steel manifold; 2 outlets; 1/4" inlet NPT(F) (YO-31521-31)
304 Stainless Steel manifold; 2 outlets; 3/8" inlet NPT(F) (YO-31521-33)
304 Stainless Steel manifold; 2 outlets; 1/2" inlet NPT(F) (YO-31521-35).
More Details
Largle Plastic Buchner Funnels
Buy Largle Plastic Buchner Funnels Items Including:
High-density polyethylene Buchner funnel, 42.5 mm dia (YO-06128-02)
High-density polyethylene Buchner funnel, 55 mm dia (YO-06128-03)
Polypropylene Buchner funnel, 110 mm dia (YO-06128-05)
Polypropylene Buchner funnel, 150 mm dia (YO-06128-10).
More Details
Phase Ii Digital Coating Thickness Gauges
Buy Phase Ii Digital Coating Thickness Gauges Items Including:
Phase II PTG-3750 Coating Thickness Gauge with external probe.
More Details
Air Cadet Vacuum Pump 07350
Buy Air Cadet Vacuum Pump 07350 Items Including:
Air Cadet® Vacuum/Pressure Pump, Diaphragm, single head, 0.8 cfm,
Air Cadet® Vacuum/Pressure Pump, Diaphragm, dual head, 0.9 cfm, 11
Air Cadet® Vacuum/Pressure Pump, Diaphragm, 0.5 cfm, 115 VAC
Nitrile diaphragm service kit for all Air Cadet pumps.
More Details
Bain Marie Containers, Hdpe
Buy Bain Marie Containers, Hdpe Items Including:
Bain Marie Container, HDPE, 12 quart
Bain Marie Container, HDPE, 16 quart
Bain Marie Container, HDPE, 20 quart
Bain Marie Container, HDPE, 24 quart
Bain Marie Container, HDPE, 32 quart
More Details
Direct Coupled Rotary Vane Pumps
Buy Direct Coupled Rotary Vane Pumps Items Including:
High-pressure, brass-bodied rotary vane pump, 0.75 GPM, 115/230 VAC
High-pressure, brass-bodied rotary vane pump, 2.3 GPM, 115/230 VAC, 5
High-pressure, 303 stainless steel rotary vane pump, 2.3 GPM, 115/230.
More Details
Gilmont Micrometer Syringe
Buy Gilmont Micrometer Syringe Items Including:
Gilmont Micrometer Syringe, 0.2 mL
Gilmont micrometer syringe tip, for use with 07840-00
Gilmont micrometer syringe, 2 mL
Gilmont micrometer syringe tip, for use with 07844-00
Gilmont micrometer burette, 2 mL.
More Details
70618
results
<<<Previous
4
5
6
7
8
9
10
11
12
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Dissolved Oxygen Meter to Gilmont Micrometer Syringe
>
31125329-s-toc_2 31125329stoc2