Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 80
Kimax Glass Centrifuge Tubes
to
Transparent Plastic Bulk Storage Bins
Image
Product Detail
More Detail
Kimax Glass Centrifuge Tubes
Buy Kimax Glass Centrifuge Tubes Items Including:
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 5 ml, 125 (99502-5)
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 10 ml, 12 (99502-10)
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 15 ml, 12 (99502-15).
More Details
Kimble Glass Centrifuge Tubes
Buy Kimble Glass Centrifuge Tubes Items Including:
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 5 ml, 125 (99502-5)
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 10 ml, 12 (99502-10)
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 15 ml, 12 (99502-15).
More Details
100ml Glass Centrifuge Tube
Buy 100ml Glass Centrifuge Tube Items Including:
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 5 ml, 125 (99502-5)
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 10 ml, 12 (99502-10)
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 15 ml, 12 (99502-15).
More Details
Pressure Gauge with Diaphragm Seal
Buy Pressure Gauge with Diaphragm Seal Items Including:
PVDF Pressure Gauge, PTFE diaphragm seal, 1/4" NPT(F); 0 to 15 psi
PVDF Pressure Gauge, PTFE diaphragm seal, 1/4" NPT(F); 0 to 30 psi
In-line Seal, 1" Process Conn, w/ 15 PSI Gauge
In-line Seal, 1" Process Conn, w/ 30 PSI Gauge.
More Details
Staining Dishes with Rack and Cover
Buy Staining Dishes with Rack and Cover Items Including:
Staining Dish, 20-Slide Capacity (YO-48585-00)
Staining Dish, 50-Slide Capacity (YO-48585-10)
Dish and Cover for 48585-00 (YO-48585-01)
Handle for 48585-00 (YO-48585-03)
Staining dish and cover (YO-48585-70)
More Details
Silicone (Peroxide Cured) Autoanalysis/Microbore Extension Tubing
Buy Silicone (Peroxide Cured) Autoanalysis/Microbore Extension Tubing Items Including:
Silicone (peroxide cured) extension tubing, 0.64 mm ID, 50 ft/pack
Silicone (peroxide cured) extension tubing, 0.76 mm ID, 50 ft/pack
Silicone (peroxide cured) extension tubing, 0.89 mm ID, 50 ft/pack.
More Details
Stainless Steel Cam Adapters and Couplers
Buy Stainless Steel Cam Adapters and Couplers Items Including:
Female Cam Adapter Cap, 1/2-in, SS (QC-DC050-SS)
Male Cam X 3/4-in NPT(M) Adapter, SS (QC-F075-SS)
Male Cam X 2-in NPT(M) Adapter, SS (QC-F200-SS)
Male Cam X 1/2-in NPT(F) Adapter, SS (QC-A050-SS)
Female Cam X 2-in NPT(M) Adapter, SS (QC-B200-SS).
More Details
Edwards 2 Inch Needle Valves
Buy Edwards 2 Inch Needle Valves Items Including:
Chemically Inert Panel-Mount Needle Valve, 1/8" NPT(M) (NV2-2-1)
Chemically Inert Panel-Mount Needle Valve, 1/4" NPT(M) (NV4-4-2)
PP Needle Valve, 1/2" NPT(F) (NV40050T)
Needle Valve, Hard Seat ss Male-Female 1/2 in
NPT Male-Female.
More Details
Battery Operated Low Pressure Vacuum Pump
Buy Battery Operated Low Pressure Vacuum Pump Items Including:
Hand-operated vacuum pump, 16 mL/stroke, PVC body
Hand-operated vacuum pump (without gauge), 32 mL/stroke, PVC body
Additel 912 Low Pressure Pneumatic Test Pump, 95% Vacuum to 60psi
Hand-operated vacuum/pressure pump, 15 mL/stroke, zinc alloy body.
More Details
Nir Turbidity Calibration Standards
Buy Nir Turbidity Calibration Standards Items Including:
Turbidity Standards Kit, linear calibration set from 0.2 to 1.0 NTU (YO-08391-26)
Turbidity Standards Kit, linear calibration set from 2.00 to 10.0 NTU (YO-08391-27)
Turbidity Standard, 10.0 NTU, 1-L bottle
Turbidity Standard, 40.0 NTU, 1-L bottle.
More Details
Procal Turbidity Calibration Standards
Buy Procal Turbidity Calibration Standards Items Including:
Turb Standard 1.0, Hach 1720
Turb Standard 0.0, Hach 2100n
Turb Standard 200, Hach 2100n
Turb Standard 1000, Hach 2100n
Turb Standard 0.0, Hach 2100p
More Details
Black Plastic Storage Bins
Buy Black Plastic Storage Bins Items Including:
Storage bin; 4-1/8" x 3" x 5-3/8", 24/pack (30-210 YELLOW)
Storage bin; 4-1/8" x 3" x 7-3/8", 24/pack (30-220 YELLOW)
Storage bin; 5-1/2" x 5" x 10-7/8", 12/pack (30-230 YELLOW).
More Details
Bulk Plastic Storage Bins 12x12x15
Buy Bulk Plastic Storage Bins 12x12x15 Items Including:
Storage bin; 4-1/8" x 3" x 5-3/8", 24/pack (30-210 YELLOW)
Storage bin; 4-1/8" x 3" x 7-3/8", 24/pack (30-220 YELLOW)
Storage bin; 5-1/2" x 5" x 10-7/8", 12/pack (30-230 YELLOW).
More Details
Christmas Plastic Storage Bins
Buy Christmas Plastic Storage Bins Items Including:
Storage bin; 4-1/8" x 3" x 5-3/8", 24/pack (30-210 YELLOW)
Storage bin; 4-1/8" x 3" x 7-3/8", 24/pack (30-220 YELLOW)
Storage bin; 5-1/2" x 5" x 10-7/8", 12/pack (30-230 YELLOW).
More Details
Color Plastic Shoe Storage Bins
Buy Color Plastic Shoe Storage Bins Items Including:
Storage bin; 4-1/8" x 3" x 5-3/8", 24/pack (30-210 YELLOW)
Storage bin; 4-1/8" x 3" x 7-3/8", 24/pack (30-220 YELLOW)
Storage bin; 5-1/2" x 5" x 10-7/8", 12/pack (30-230 YELLOW).
More Details
Colored Plastic Storage Bins
Buy Colored Plastic Storage Bins Items Including:
Storage bin; 4-1/8" x 3" x 5-3/8", 24/pack (30-210 YELLOW)
Storage bin; 4-1/8" x 3" x 7-3/8", 24/pack (30-220 YELLOW)
Storage bin; 5-1/2" x 5" x 10-7/8", 12/pack (30-230 YELLOW).
More Details
Commercial Plastic Storage Bins
Buy Commercial Plastic Storage Bins Items Including:
Storage bin; 4-1/8" x 3" x 5-3/8", 24/pack (30-210 YELLOW)
Storage bin; 4-1/8" x 3" x 7-3/8", 24/pack (30-220 YELLOW)
Storage bin; 5-1/2" x 5" x 10-7/8", 12/pack (30-230 YELLOW).
More Details
Covered Plastic Storage Bins
Buy Covered Plastic Storage Bins Items Including:
Storage bin; 4-1/8" x 3" x 5-3/8", 24/pack (30-210 YELLOW)
Storage bin; 4-1/8" x 3" x 7-3/8", 24/pack (30-220 YELLOW)
Storage bin; 5-1/2" x 5" x 10-7/8", 12/pack (30-230 YELLOW).
More Details
Epic Plastic Storage Bins
Buy Epic Plastic Storage Bins Items Including:
Storage bin; 4-1/8" x 3" x 5-3/8", 24/pack (30-210 YELLOW)
Storage bin; 4-1/8" x 3" x 7-3/8", 24/pack (30-220 YELLOW)
Storage bin; 5-1/2" x 5" x 10-7/8", 12/pack (30-230 YELLOW).
More Details
Jumbo Plastic Storage Bins
Buy Jumbo Plastic Storage Bins Items Including:
Storage bin; 4-1/8" x 3" x 5-3/8", 24/pack (30-210 YELLOW)
Storage bin; 4-1/8" x 3" x 7-3/8", 24/pack (30-220 YELLOW)
Storage bin; 5-1/2" x 5" x 10-7/8", 12/pack (30-230 YELLOW).
More Details
Large Clear Plastic Storage Bins
Buy Large Clear Plastic Storage Bins Items Including:
Storage bin; 4-1/8" x 3" x 5-3/8", 24/pack (30-210 YELLOW)
Storage bin; 4-1/8" x 3" x 7-3/8", 24/pack (30-220 YELLOW)
Storage bin; 5-1/2" x 5" x 10-7/8", 12/pack (30-230 YELLOW).
More Details
Plastic Book Storage Bins
Buy Plastic Book Storage Bins Items Including:
Storage bin; 4-1/8" x 3" x 5-3/8", 24/pack (30-210 YELLOW)
Storage bin; 4-1/8" x 3" x 7-3/8", 24/pack (30-220 YELLOW)
Storage bin; 5-1/2" x 5" x 10-7/8", 12/pack (30-230 YELLOW).
More Details
Plastic Clothing Storage Bins
Buy Plastic Clothing Storage Bins Items Including:
Storage bin; 4-1/8" x 3" x 5-3/8", 24/pack (30-210 YELLOW)
Storage bin; 4-1/8" x 3" x 7-3/8", 24/pack (30-220 YELLOW)
Storage bin; 5-1/2" x 5" x 10-7/8", 12/pack (30-230 YELLOW).
More Details
Plastic Storage Bins False Office
Buy Plastic Storage Bins False Office Items Including:
Storage bin; 4-1/8" x 3" x 5-3/8", 24/pack (30-210 YELLOW)
Storage bin; 4-1/8" x 3" x 7-3/8", 24/pack (30-220 YELLOW)
Storage bin; 5-1/2" x 5" x 10-7/8", 12/pack (30-230 YELLOW).
More Details
Quantam Plastic Storage Bins
Buy Quantam Plastic Storage Bins Items Including:
Storage bin; 4-1/8" x 3" x 5-3/8", 24/pack (30-210 YELLOW)
Storage bin; 4-1/8" x 3" x 7-3/8", 24/pack (30-220 YELLOW)
Storage bin; 5-1/2" x 5" x 10-7/8", 12/pack (30-230 YELLOW).
More Details
Round Plastic Storage Bins
Buy Round Plastic Storage Bins Items Including:
Storage bin; 4-1/8" x 3" x 5-3/8", 24/pack (30-210 YELLOW)
Storage bin; 4-1/8" x 3" x 7-3/8", 24/pack (30-220 YELLOW)
Storage bin; 5-1/2" x 5" x 10-7/8", 12/pack (30-230 YELLOW).
More Details
Storage Bins Containers
Buy Storage Bins Containers Items Including:
Storage bin; 4-1/8" x 3" x 5-3/8", 24/pack (30-210 YELLOW)
Storage bin; 4-1/8" x 3" x 7-3/8", 24/pack (30-220 YELLOW)
Storage bin; 5-1/2" x 5" x 10-7/8", 12/pack (30-230 YELLOW).
More Details
Storage Bins Plastic 66 Quart
Buy Storage Bins Plastic 66 Quart Items Including:
Storage bin; 4-1/8" x 3" x 5-3/8", 24/pack (30-210 YELLOW)
Storage bin; 4-1/8" x 3" x 7-3/8", 24/pack (30-220 YELLOW)
Storage bin; 5-1/2" x 5" x 10-7/8", 12/pack (30-230 YELLOW).
More Details
Tall Plastic Storage Bins
Buy Tall Plastic Storage Bins Items Including:
Storage bin; 4-1/8" x 3" x 5-3/8", 24/pack (30-210 YELLOW)
Storage bin; 4-1/8" x 3" x 7-3/8", 24/pack (30-220 YELLOW)
Storage bin; 5-1/2" x 5" x 10-7/8", 12/pack (30-230 YELLOW).
More Details
Transparent Plastic Bulk Storage Bins
Buy Transparent Plastic Bulk Storage Bins Items Including:
Storage bin; 4-1/8" x 3" x 5-3/8", 24/pack (30-210 YELLOW)
Storage bin; 4-1/8" x 3" x 7-3/8", 24/pack (30-220 YELLOW)
Storage bin; 5-1/2" x 5" x 10-7/8", 12/pack (30-230 YELLOW).
More Details
70618
results
<<<Previous
76
77
78
79
80
81
82
83
84
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
20
30
40
50
60
65
66
67
68
69
70
71
72
73
74
75
76
77
78
79
80
81
82
83
84
85
86
87
88
89
90
91
92
93
94
100
110
120
130
140
150
160
170
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Kimax Glass Centrifuge Tubes to Transparent Plastic Bulk Storage Bins
>
31125329-s-toc_2 31125329stoc2