Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 800
General Tools Humidity Indicators
to
Brass Male Compression Elbow
Image
Product Detail
More Detail
General Tools Humidity Indicators
Buy General Tools Humidity Indicators Items Including:
Digital Termperature and Humidity Monitor with Clock and Alarm.
More Details
Platinum Silicone Tubing
Buy Platinum Silicone Tubing Items Including:
Platinum-Cured Silicone Tubing, 3/32"ID x 5/32"OD, 25 ft/pack
Platinum-Cured Silicone Tubing, 5/32"ID x 7/32"OD, 25 ft/pack
Platinum-Cured Silicone Tubing, 3/16"ID x 5/16"OD, 25 ft/pack.
More Details
Quick Coupling
Buy Quick Coupling Items Including:
Ismatec™ quick-couple digital drive, 2 to 240 rpm, 115/230 VAC
Ismatec™ quick-change pump head, up to 2810 mL/min, nominal tubing
Ismatec™ quick-change pump head, up to 876 mL/min, nominal tubing
Coupling Body, Valved, 1/4" OD Tube (PMCD1304).
More Details
Tefzel Accessories for Liquid Chromatography (Lc)
Buy Tefzel Accessories for Liquid Chromatography (Lc) Items Including:
Flangeless Ferrule, 1/16", Tefzel, Pack Of 10 (YO-02007-54).
More Details
Whatman Accessories for Liquid Chromatography (Lc)
Buy Whatman Accessories for Liquid Chromatography (Lc) Items Including:
Whatman In-Line Solvent Degasser, Aqueous Phase; 10/box.
More Details
Cole-Parmer Mixing Drum Pumps
Buy Cole-Parmer Mixing Drum Pumps Items Including:
Mixing drum pump, 24 GPM, PP
Pump and mix tube set, 47", Stainless Steel (F426S 41/38 47")
Pump and mix tube set, 60", Stainless Steel (F426S 41/38 60")
Sanitary Batch Control System, 3A, 110 VAC/50Hz, 1HP TEFC, 3 GPM (11 L.
More Details
Ph Conductivity and Resistivity Meter
Buy Ph Conductivity and Resistivity Meter Items Including:
MYRON L 4PII Waterproof Conductivity/TDS/Resistivity Meter
Hach® sensION™ 5 Waterproof Conductivity Meter
MYRON L TechPro II™ TP1 conductivity/TDS meter
Oakton waterproof CON 610 handheld conductivity/TDS/salinity/resistivi (CON 610) (YO-35408-10).
More Details
Reaction Flask Heating Mantle
Buy Reaction Flask Heating Mantle Items Including:
Glas-Col aluminum heating mantle, 1000-ml, 380 watts, 115 VAC
Glas-Col aluminum heating mantle, 1000-ml, 380 watts, 230 VAC
Glas-Col aluminum heating mantle, 2000-ml, 500 watts, 115 VAC
Glas-Col aluminum heating mantle, 2000-ml, 500 watts, 230 VAC.
More Details
Cole-Parmer Pony Utility Pumps
Buy Cole-Parmer Pony Utility Pumps Items Including:
Pony Utility Pump for Intermittent Use, Nickel Plated Brass, 110VAC
Pony Utility Pump for Intermittent Use, Nickel Plated Brass, 12VDC
Pump, Utility Accessory Hose kit consisting of 10' garden hose, SST sp.
More Details
Ozone Gas Detectors
Buy Ozone Gas Detectors Items Including:
Digital ozone monitor
Ozone monitor with alarm
Continuous duty ozone monitor.
More Details
Stainless Steel Vacuum Filtration Manifolds
Buy Stainless Steel Vacuum Filtration Manifolds Items Including:
Advantec Vacuum Filtration Manifold; Stainless Steel, 3-Branch with 2-
Advantec Stainless Steel Pressure Filtration Holder with Reservoir, fo (YO-02926-32)
Advantec Stainless Steel Pressure Filtration Holder with Reservoir, fo (YO-02926-54).
More Details
Variable Speed Pump Drives
Buy Variable Speed Pump Drives Items Including:
Masterflex® I/P® Variable Speed Modular Drive, 20-650 RPM, 230 (07591-07)
Series Q single head, variable-speed AC drive with controller, 1800 sp (QV-0710432)
Masterflex® L/S® variable-speed drive with 10-turn speed contr (77521-57).
More Details
Bsp Threaded Pipe Fittings
Buy Bsp Threaded Pipe Fittings Items Including:
Pipe fittings, NPT(F) x BSPT(M) adapter, BR, 1/4", 6200 psi, 1-3/8", (YO-31834-01).
More Details
Carbon Dioxide Incubator Gas
Buy Carbon Dioxide Incubator Gas Items Including:
Carbon Dioxide Traps; 40 liters volume; 1/8" fittings (YO-34651-60)
Carbon Dioxide Traps; 40 liters volume; 1/4" fittings (YO-34651-70)
Sensidyne Gas Detection Tubes, Carbon dioxide
Sensidyne Gas Detection Tubes, Carbon Dioxide, 5 to 50%.
More Details
Female to Hose Barb
Buy Female to Hose Barb Items Including:
Miniature Quick-Disconnect Hose Barb (Female); Valved, 1/8" ID (SMFD02)
Female Cam X 3/4-in Hose Barb Adapter, PP (QC-C075-PP)
Female Cam X 1-1/2-in Hose Barb Adapter, PP (QC-C150-PP)
Female Cam X 1/2-in Hose Barb Adapter, PVDF (QC-C050-PVDF).
More Details
Dc Air Pumps
Buy Dc Air Pumps Items Including:
Series Q single head, enclosed DC drive, 1800 spm, 12 VDC (QB-0710420)
Series Q single head, enclosed DC drive, 1800 spm, 24 VDC (QB 0710425)
Series Q single head, air powered drive, 400 spm (GPD).
More Details
Wl Gore Silicone Peristaltic Pump Tubing
Buy Wl Gore Silicone Peristaltic Pump Tubing Items Including:
Masterflex® peroxide-cured silicone tubing, I/P® 89, 10 ft
(YO-96400-89)
L/S Digital Pump System with GORE High-Resilience Tubing Style 500 and.
More Details
High Power Dc Supply
Buy High Power Dc Supply Items Including:
B&K Precision 1901 DC Switching Power Supply, 960W
B&K Precision 1902 DC Switching Power Supply, 900W
B&K Precision 1900 DC Switching Power Supply, 960W
Instek Model GPD-3303D, Multi Output, High Resolution, Programmable Li (GPD-3303D).
More Details
Sheet Metal Lab Fume Hood
Buy Sheet Metal Lab Fume Hood Items Including:
Organic Vapor Replacement Filters for Portable Ductless Fume Hoods (MF4AS)
Acid Vapor Replacement Filters for Portable Ductless Fume Hoods (MF4BE)
Prefilter for 24" and 36" fume hoods (FE-2013)
Prefilter for 48" fume hoods (FE-3013).
More Details
Amber Glass Media Bottles
Buy Amber Glass Media Bottles Items Including:
30 mL Amber Glass Dropping Bottle, Plastic dropper
Amber glass dropping bottle, 120 mL
Pack of 6
(W211755)
500 mL Kimax Amber Media Bottles
More Details
Cole-Parmer Pressure Booster Pump Systems
Buy Cole-Parmer Pressure Booster Pump Systems Items Including:
Pump Booster System, 38 GPM or 45 PSI, 1 HP, NEMA 1 enclosure
Pump Booster System, 38 GPM or 63 PSI, 2 HP, NEMA 1 enclosure
Pump Booster System, 60 GPM or 33 PSI, 1 HP, NEMA 1 enclosure
Pump Booster System, 60 GPM or 63 PSI, 3 HP, NEMA 1 enclosure.
More Details
Hypalon Positive Displacement Pumps
Buy Hypalon Positive Displacement Pumps Items Including:
Large-Hose Peristaltic Pump, Lubricant-Filled Pump Cavity, Hypalon Hos (3005-HF-PV-F2)
Large-Hose Peristaltic Pump, Dry Pump Cavity, Hypalon Hose, 57 GPM, co
Large-Hose Peristaltic Pump, Lubricant-Filled Pump Cavity, Hypalon Hos (3005-HF-PV-J2).
More Details
Polyolefin Heat Shrink Tubing
Buy Polyolefin Heat Shrink Tubing Items Including:
Polyolefin heat-shrink tubing, 0.125 (YO-06850-10)
Polyolefin heat-shrink tubing, 0.25 (YO-06850-20)
Polyolefin heat-shrink tubing, 0.375 (YO-06850-30)
Polyolefin heat-shrink tubing, 0.50 (YO-06850-40)
Tubing,heat-shrink,3/64, 25' (YO-95219-00).
More Details
Liquid Filled Pressure Gauges
Buy Liquid Filled Pressure Gauges Items Including:
Wika Forged Brass Industrial Gauge ~ 0 To 5000 PSI, 2 1/2"
Wika Forged Brass Industrial Gauge ~ 0 To 1000 PSI, 2 1/2"
Wika Forged Brass Industrial Gauge ~ 0 To 3000 PSI, 2 1/2"
Wika Forged Brass Industrial Gauge 4in 30inhg 1/2 l.
More Details
Soxhlet Flask
Buy Soxhlet Flask Items Including:
Soxhlet 125 ml flat bottom flask (YO-99037-44)
Soxhlet 250 ml flat bottom flask (YO-99037-46)
Soxhlet 125 ml round bottom flask (YO-99037-60)
Soxhlet 250 ml round bottom flask (YO-99037-62)
Soxhlet 300 ml round bottom flask (YO-99037-64).
More Details
Water Quality Testing Lab
Buy Water Quality Testing Lab Items Including:
Refill kit, Phenol test, for 00290-30
Refill kit, Copper, total soluble test, for 00290-50
Refill kit, Phosphate test, for 00290-52
Refill kit, Iron, total soluble test, for 00291-40
Replacement reagents for basic soil chemistry test kit (R-5029).
More Details
5 Gallon Container with Spigot
Buy 5 Gallon Container with Spigot Items Including:
FDA-approved high-density polyethylene container, 5 gallon (HPB 4716)
Collapsible Container, Low-Density Polyethylene (LDPE), 5 gallon
Box, HDPE, 5 gallon
Calibration container
HDPE, 5-gallon capacity
More Details
Process Gauges
Buy Process Gauges Items Including:
Ashcroft Duragauge Plus! Process Gauge, 30" Hg to 15 psi
Ashcroft Duragauge Plus! Process Gauge, 30" Hg to 30 psi
Ashcroft Duragauge Plus! Process Gauge, 0 to 1000 psi
Ashcroft Duragauge Plus! Process Gauge, 0 to 3000 psi.
More Details
Cole-Parmer Fittings for Tubing and Hose
Buy Cole-Parmer Fittings for Tubing and Hose Items Including:
Cole-Parmer, Threaded Sanitary Clamp Adapter, 316L SS, 1.5" Clamp X 3/
Barbed fitting kits, Barbed connector/reducer kits, PP, small fittings (YO-31514-03)
Barbed fitting kits, Threaded barb adapter kits, Natural PP, small fit (YO-31514-11).
More Details
Brass Male Compression Elbow
Buy Brass Male Compression Elbow Items Including:
Compression elbow union, 1/8", Brass, each (YO-31412-06)
Compression elbow union, 1/4", Brass, each (YO-31412-07)
Compression elbow union, 3/8", Brass, each (YO-31412-08)
Compression elbow union, 1/2", Brass, each (YO-31412-09).
More Details
70618
results
<<<Previous
796
797
798
799
800
801
802
803
804
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
100
200
300
400
500
600
700
710
720
730
740
750
760
770
780
785
786
787
788
789
790
791
792
793
794
795
796
797
798
799
800
801
802
803
804
805
806
807
808
809
810
811
812
813
814
820
830
840
850
860
870
880
890
900
1,000
1,100
1,200
1,300
1,400
1,500
1,600
1,700
2,000
2,354
General Tools Humidity Indicators to Brass Male Compression Elbow
>
31125329-s-toc_2 31125329stoc2