Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Product Index Page 90
Magnetic Thermocouple Probe
to
Temperature Systems and Probe only Calibrations
Image
Product Detail
More Detail
Magnetic Thermocouple Probe
Buy Magnetic Thermocouple Probe Items Including:
Type T Dropping/magnetic thermocouple probe; 1.5L (072197A-T)
Type K Dropping/magnetic thermocouple probe; 1.5"L (113001B-K)
Type J Dropping/magnetic thermocouple probe; (113001B-J)
Type T Dropping/magnetic thermocouple probe (113001B-T).
More Details
Reusable Temperature Labels / Indicators
Buy Reusable Temperature Labels / Indicators Items Including:
Temperature Labels, Refrigerator (YO-90308-80)
HOT Hand Reversible Indicating Label, Changes from orange to yellow at (4071COPADP10)
HOT Hand Reversible Indicating Label, Changes from Red to black at 157 (4077COPADP10).
More Details
Brushless Dc Magnetic Drive Centrifugal Pumps
Buy Brushless Dc Magnetic Drive Centrifugal Pumps Items Including:
Brushless DC Magnetic Drive Pump, Polypropylene, 3.5 GPM, 12 VDC
Brushless DC Magnetic Drive Pump, Polypropylene, 8 GPM, 12 VDC
Brushless DC Magnetic Drive Pump, Polypropylene, 8 GPM, 24 VDC
Brushless DC Magnetic Drive Pump, PVDF, 3.5 GPM, 24 VDC.
More Details
Reference Electrode Filling Solution
Buy Reference Electrode Filling Solution Items Including:
ROSS electrode fill solution, 3 M KCl
Case of five 60-mL bottles (810007)
Thermo Scientific Orion electrode fill solution, 3 M KCl, 5 x 60mL bot
Filling solutions: use with Cl- electrodes (900017)
Filling solution f/ammonia ISE electrode (951202).
More Details
Small Sample Containers with Seperate Lid
Buy Small Sample Containers with Seperate Lid Items Including:
Hinged-Lid Sample Containers, PP, 1/4 oz
Hinged-Lid Sample Containers, PP, 1/2 oz
Hinged-Lid Sample Containers, PP, 3/4 oz
Hinged-Lid Sample Containers, PP, 1 oz
Hinged-Lid Sample Containers, PP, 2 oz
More Details
3 Hp Vacuum Pumps
Buy 3 Hp Vacuum Pumps Items Including:
Cole-Parmer® Two Stage Rotary Vane Vacuum Pump, 106/128L/min, 3x10 (YO-72226-65)
Vacuubrandreg Rotary Vane Vacuum Pump, 1.6 cfm, 1.5 X 10-3/sup (698123)
Vacuubrandreg Rotary Vane Vacuum Pump, 1.6 cfm, 1.5 X 10-3/sup (698120).
More Details
Thermo Scientific Barnstead Water Deionization Systems
Buy Thermo Scientific Barnstead Water Deionization Systems Items Including:
Thermo Scientific Barnstead Ultrapure deionization cartridge, 680 grai (D50253)
Thermo Scientific Barnstead Pretreatment Cartridge for tap and deioniz (D50251)
Thermo Scientific Barnstead GenPure UF/UV
Thermo Scientific Barnstead GenPure UF/UV TOC.
More Details
Heidolph Oem Vacuum Pump from Who
Buy Heidolph Oem Vacuum Pump from Who Items Including:
Compact/oem Vacuum Pump, 12 VDC, 1.4 LPM, 8.8
PUMP, VACUUM PUMP WITH BRUSHLESS DC MOTOR, 1.5 LPM, 12 VDC, GAS ONLY,
Rotary vane oil vacuum pump, 3.75 cfm, 220 VAC (0523-V4J-G588DX)
Dry rotary vane vacuum pump, 4.5 cfm, 115 VAC (0523-V542AQ-G588DX).
More Details
Handles for Oval Brushes
Buy Handles for Oval Brushes Items Including:
Handle for 84550-series oval tank brushes, 42" long.
More Details
Ph Test Strips Chart
Buy Ph Test Strips Chart Items Including:
EM Quant pH test strips, 0-14 pH
Six packs of 100.
pHydrion plastic pH indicator strips, 0.0 to 14.0, flip top vial packa
Universal pH Test Paper; pH Range 1 to 11; 9 mm x 6 m Roll
Whole Range pH Test Paper; pH Range 1 to 14; 9 mm x 5.5 m Roll.
More Details
Tygon Silicone Tubing
Buy Tygon Silicone Tubing Items Including:
Tygon Silicone Tubing, 1/32"ID x 3/32"OD, 50 ft/pack
Tygon Silicone Tubing, 1/16"ID x 1/8"OD, 50 ft/pack
Tygon Silicone Tubing, 1/8"ID x 3/16"OD, 50 ft/pack
Tygon Silicone Tubing, 1/8"ID x 1/4"OD, 50 ft/pack.
More Details
Stainless Steel Female/Male Manual Ball Valves
Buy Stainless Steel Female/Male Manual Ball Valves Items Including:
1/4" NPT stainless steel female/male manual ball valve (XV501SS-4)
3/8" NPT stainless steel female/male manual ball valve (XV501SS-6)
3/4" NPT stainless steel female/male manual ball valve (XV501SS-12).
More Details
Industrial Centrifugal Pumps
Buy Industrial Centrifugal Pumps Items Including:
Pump Less Volute, Centrifugal pump, for either all bronze or bronze fi (YO-79730-56)
Pump Less Volute, Centrifugal pump, for either all bronze or bronze fi (YO-79730-57)
Pump, Centrifugal Pump, Hot water recirculation pump, 6 GPM, 3/4" Swea.
More Details
Other John Guest Acetal Push-to-Connect Fittings
Buy Other John Guest Acetal Push-to-Connect Fittings Items Including:
Push-to-connect check valve, 3/8", 10/pack (YO-34006-64)
Push-to-connect plugs, 3/16", 10/pack (YO-34006-93)
Push-to-connect collet covers, 3/16", 10/pack (YO-34006-95)
Push-to-connect collet covers, 1/4", 10/pack (YO-34006-96).
More Details
Ultra-Quiet Air Compressors
Buy Ultra-Quiet Air Compressors Items Including:
Ultra-Quiet Oil-lubricated Air Compressor, 0.5 cfm, 115 VAC (P15-TC)
Ultra-Quiet Oil-lubricated Air Compressor, 1.5 cfm, 115 VAC (P50/24AL)
Ultra-Quiet Oil-lubricated Air Compressor, 4.5 cfm, 115 VAC (P150/50AL).
More Details
High-Efficiency Polyester Absolute-Rated Filter Bags
Buy High-Efficiency Polyester Absolute-Rated Filter Bags Items Including:
Size 1 high-efficiency polyester absolute-rated filter bag; 300°F (255120-03)
Size 1 high-efficiency polyester absolute-rated filter bag; 300°F (255122-03)
Size 1 high-efficiency polyester absolute-rated filter bag; 300°F (255124-03).
More Details
Product Type Button Cell Batteries
Buy Product Type Button Cell Batteries Items Including:
Button cell batteries, 1.5 V, 6/pack (Eveready # 386) (386BP)
Button cell batteries, 1.5 V, 6/pack (Eveready # 392) (392BP)
Button cell batteries, 1.5 V, 5/pack (Eveready # 393) (393)
Button cell batteries, 1.5 V, 6/pack (Eveready # 389BP) (389BP).
More Details
Thermo Scientific Nalgene 175-Ml Centrifuge Bottles
Buy Thermo Scientific Nalgene 175-Ml Centrifuge Bottles Items Including:
Thermo Scientific Nalgene Centrifuge bottle; 175-mL; PPCO; 4/pack.
More Details
Omega Digital Pressure Gauge Gauges
Buy Omega Digital Pressure Gauge Gauges Items Including:
Digital Pressure Gauge 0 to 800 psi
Digital Pressure Gauge, 0-100 PSI
Digital Pressure Gauge, 0-200 PSI
Digital Pressure Gauge, 0-300 PSI
Digital Pressure Gauge, 0-500 PSI
More Details
Chemical Resistant Trays
Buy Chemical Resistant Trays Items Including:
Chemical Resistant Tray - 13-1/4" X 10-5/8", 1 Ea (3034031537)
Chemical Resistant Tray - 20-3/8" X 15-1/8", 1 Ea (3044031537)
Chemical Resistant Pan - 25-1/4" X 19-1/4", 1 Ea (6061031537)
Chemware Tray PFA, With Cover.
More Details
Mortar and Pestle
Buy Mortar and Pestle Items Including:
Cole-Parmer Mortar and Pestle Set, Porcelain; 80ML, Each
Porcelain Pestle Grinding Set (YO-04181-51)
Agate Pestle Grinding Set (YO-04181-61)
Hardened Chrome Steel Pestle Grinding Set (YO-04181-63)
Stainless Steel Pestle Grinding Set (YO-04181-67).
More Details
Hayward Valves
Buy Hayward Valves Items Including:
Hayward True Union Ball Check Valve, 1/2" socket and NPT(F) connectors
Hayward True Union Ball Check Valve, 3/4" socket and NPT(F) connectors
Hayward Electrically Actuated PVC True Union Ball Valve, 1/2", 115 VAC.
More Details
Sperian Atlas Fit Rubber Dipped Gloves
Buy Sperian Atlas Fit Rubber Dipped Gloves Items Including:
Sperian® Atlas Fit™ Rubber Coated Cotton/Poly Glove, Medium
Atlas Fit® Rubber Coated Cotton Poly Glove Black and Gray Sm (320-S)
Atlas Fit® Rubber Coated Cotton Poly Glove Black and Gray Lg (320-L).
More Details
Water Softening Systems
Buy Water Softening Systems Items Including:
Compact water softening system with timeclock regeneration and top-mou (162404 (W-427F))
Compact water softening system with meter regeneration and top-mount v (162401 (W-425F)).
More Details
Cole One-Way Luer Check Valves
Buy Cole One-Way Luer Check Valves Items Including:
One-way luer check valve, SAN/LDPE with silicone diaphragm, female-mal (SCV08053).
More Details
Oilless Air Compressors with Storage Tanks
Buy Oilless Air Compressors with Storage Tanks Items Including:
Oilless Air Compressor with 20 Gallon Storage Tank, 3/4 hp, 115 VAC/60 (5HCD-11T-M550NGX)
Oilless Air Compressor with 20 Gallon Storage Tank, 3/4 hp, 230 VAC/60 (5HCD-11TA-M550X).
More Details
Arm Length Disposable Nitrile Gloves
Buy Arm Length Disposable Nitrile Gloves Items Including:
Best Nighthawk™ Black Nitrile Gloves, Small, Box of 50
Kimberly-Clark Nitrile Gloves, 9.5" length, Small, 100/box
Kimberly-Clark Nitrile Gloves, 9.5" length, Medium; 100/box
Kimberly-Clark Nitrile Gloves, 9.5" length, Large; 100/box.
More Details
Elbow Length Disposable Nitrile Gloves
Buy Elbow Length Disposable Nitrile Gloves Items Including:
Best Nighthawk™ Black Nitrile Gloves, Small, Box of 50
Kimberly-Clark Nitrile Gloves, 9.5" length, Small, 100/box
Kimberly-Clark Nitrile Gloves, 9.5" length, Medium; 100/box
Kimberly-Clark Nitrile Gloves, 9.5" length, Large; 100/box.
More Details
Sanitary Clamp End Cap
Buy Sanitary Clamp End Cap Items Including:
316L Stainless Steel Asme-bpe Sanitary Clamp End Cap, 15ra Electropoli (YO-30607-29)
316L Stainless Steel Asme-bpe Sanitary Clamp End Cap, 15ra Electropoli (YO-30607-30)
End cap, PFA, for 1/4, 1/2, 3/4" sanitary clamp (YO-31530-34).
More Details
Temperature Systems and Probe only Calibrations
Buy Temperature Systems and Probe only Calibrations Items Including:
NIST-Traceable Calibration; PRT Probe (1700111)
NIST-Traceable Calibration; Thermocouple Meter+Probe System (cryogenic (1710325)
NIST-Traceable Calibration; HART TEMP RTD Meter & Probe System
NIST-Traceable Calibration; Thermocouple Probe Only.
More Details
70936
results
<<<Previous
86
87
88
89
90
91
92
93
94
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
20
30
40
50
60
70
75
76
77
78
79
80
81
82
83
84
85
86
87
88
89
90
91
92
93
94
95
96
97
98
99
100
101
102
103
104
110
120
130
140
150
160
170
180
200
300
400
500
600
700
800
900
1,000
2,000
2,365
Magnetic Thermocouple Probe to Temperature Systems and Probe only Calibrations
>
31125329-s-toc_2 31125329stoc2