Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 900
Bel-a Safety Labeled Wide-Mouth Wash Bottles
to
Weatherlink Weather Stations
Image
Product Detail
More Detail
Bel-a Safety Labeled Wide-Mouth Wash Bottles
Buy Bel-a Safety Labeled Wide-Mouth Wash Bottles Items Including:
Scienceware safety labeled 500-mL wash bottle assortment pack
Scienceware safety labeled 500-mL wash bottle for 70% ethanol
Scienceware safety labeled 500-mL wash bottle for deionized water
Scienceware safety labeled 500-mL wash bottle for acetone.
More Details
12 Volt Dc Bronze Gear Pump
Buy 12 Volt Dc Bronze Gear Pump Items Including:
Bronze Gear Pump Head, Close-Coupled Design, 2.1GPM at 0 PSI when used (YO-70738-12).
More Details
Inch Nylon Tubing Clamps
Buy Inch Nylon Tubing Clamps Items Including:
Cole Parmer White Nylon Hose Clamp Kit
Cole Parmer Black Nylon Hose Clamp Kit
Cole-Parmer Black Nylon Hose Clamps, 0.283" x 0.312" 100/pk
Cole-Parmer Black Nylon Hose Clamps, 0.350" x 0.395" 100/pk.
More Details
Split Nylon Tubing Clamps
Buy Split Nylon Tubing Clamps Items Including:
Cole Parmer White Nylon Hose Clamp Kit
Cole Parmer Black Nylon Hose Clamp Kit
Cole-Parmer Black Nylon Hose Clamps, 0.283" x 0.312" 100/pk
Cole-Parmer Black Nylon Hose Clamps, 0.350" x 0.395" 100/pk.
More Details
Handheld Mini 365 Nm Uv Lamp
Buy Handheld Mini 365 Nm Uv Lamp Items Including:
Handheld Mini 254 nm Ultraviolet Lamp (95-0158-04(UVG-4))
Standard Pen-Ray UV lamp; 365 nm, 2.25"
Standard Pen-Ray UV lamp; 254 nm, 2.13"
Standard Pen-Ray UV lamp; 254 nm, 9"
Battery-operated UV lamp, 365 nm wavelength (95-0011-01).
More Details
Laboratory Grade Gloves
Buy Laboratory Grade Gloves Items Including:
Best N-Dex Laboratory Grade Gloves, low-powder, medium
Best N-Dex Laboratory Grade Gloves, low-powder, X-large
Best N-Dex Laboratory Grade Gloves, powder-free, small
Best N-Dex Laboratory Grade Gloves, powder-free, medium.
More Details
Dual 1 4 Id Rubber Tubing
Buy Dual 1 4 Id Rubber Tubing Items Including:
Black Rubber Vacuum Tubing, 3/4" ID x 1 1/2" OD, 25ft
(YO-96143-16)
Black Rubber Vacuum Tubing, 3/4" ID x 1 1/2" OD, 25ft
(YO-96143-17)
Black Rubber Vacuum Tubing, 1/2" ID x 1 1/8" OD, 25ft
More Details
Molded Plastics Polypropylene Filters
Buy Molded Plastics Polypropylene Filters Items Including:
Thermo Scientific Nalgene polypropylene filtering flask, 500 mL
Thermo Scientific Nalgene polypropylene filtering flask, 1000 mL.
More Details
Precision General Accessories and Replacement Items
Buy Precision General Accessories and Replacement Items Items Including:
Info-con Precision Industrial RTD Probe, 9" Length, -200 To 450°C, (YO-93202-05)
HART High-Precision Platinum RTD 1/4 X 12" probe (Secon) (YO-93202-55)
Precision Industrial Thermistor probe, 9" length, 0 to 100°C, Accu (YO-93202-45).
More Details
Glentronics Float Switch Controller Bwc1
Buy Glentronics Float Switch Controller Bwc1 Items Including:
Float Switch with Piggyback Plug
Hinged float switch; nylon, 1/2" NPT external mounting (140620)
Hinged Float Switch; stainless steel, explosion proof, 1/2" x 1/2" con (M5910)
Vertical float switch; polypropylene/silicone (VS803-21).
More Details
Cell Culture Media Plates
Buy Cell Culture Media Plates Items Including:
INTEGRA Biosciences CELLine™ system, CL350, one 350-mL media reserv (YO-13052-10).
More Details
Stainless Steel Shafting
Buy Stainless Steel Shafting Items Including:
I/P long-shaft Easy-Load pump head, PSF housing, SS rotor..
More Details
Jiffy-Jack Clamps, Supports, Laboratory Jacks
Buy Jiffy-Jack Clamps, Supports, Laboratory Jacks Items Including:
Jiffy-Jack Lab Jack, 66 lb/30 kg limit
Jiffy-Jack Lab Jack, Heavy-Duty Apparatus Positioners, 20lb (9.0kg) li
Jiffy-Jack Lab Jack, 132 lb/60 kg limit
Jiffy-Jack Lab Jack, 176 lb/80 kg limit
Jiffy-Jack Lab Jack, 198 lb/90 kg limit.
More Details
Female Luer Slip
Buy Female Luer Slip Items Including:
Luer fittings, 316 SS fittings, female luer x male slip (YO-31507-20)
Large-bore adapter luer-style female x 3/16" hose barb, polycarbonate, (YO-45509-56)
Large-bore adapter luer-style female x 1/4" hose barb, polycarbonate, (YO-45509-58).
More Details
Calibration Gas Regulator
Buy Calibration Gas Regulator Items Including:
Regulator for Calibration Gas
Demand Flow Regulator for Calibration Gas
BW Technologies Demand Flow Regulator for Microdock II Base Station
BW Technologies Demand Flow Regulator with CGA connector for refillabe.
More Details
Cole-Parmer Metal Detectable Sanitary Gaskets
Buy Cole-Parmer Metal Detectable Sanitary Gaskets Items Including:
X-Rayable and Metal-Detectable Gasket, Platinum-Cured Silicone, 3/4"
X-Rayable and Metal-Detectable Gasket, Platinum-Cured Silicone, 1".
More Details
Suction Pump Accessories
Buy Suction Pump Accessories Items Including:
Drum safety wand/suction tube straightener, 2" NPT bung fitting, for s (10783)
SUCTION LINE STRAINER, PVC, 1/4" Female Friction Fit, 40 MESH (stainle
SUCTION LINE STRAINER, PVC, 1/4" Female Friction Fit, 80 MESH (stainle.
More Details
Product Type Carrying Case
Buy Product Type Carrying Case Items Including:
YSI Soft-Sided Carrying Case for Professional Series Meters
YSI Hard-Sided Carrying Case for Professional Series Meters
Carrying case with foam insert
Ohaus Carrying Case for CS/CL Scales
Hard Carrying Case for 4 1/2" and 6" Dial Gauges.
More Details
Labtek Chamber Slide Systems
Buy Labtek Chamber Slide Systems Items Including:
Lab-Tek™ II Chamber Slide™ System; material, glass; number of we (YO-01838-00)
Lab-Tek™ II Chamber Slide™ System; material, glass; number of we (YO-01838-01)
Lab-Tek™ II Chamber Slide™ System; material, glass; number of we (YO-01838-02).
More Details
Explosion Proof Transfer Pumps
Buy Explosion Proof Transfer Pumps Items Including:
Explosion-proof vacuum pump, 1.2 cfm, 27" Hg, 115 VAC (N035 STI-EX)
ATEX Explosion Proof Vacuum Pump, PVDF Coated, 7.5 l/min, 140 mbars, 1 (N87 TTE-115V)
ATEX Explosion Proof Vacuum Pump, PVDF coated, 13 l/min, 125 mbars, 1
(N726 FT.29E-115V).
More Details
General Tools & Instruments Rain Gauges
Buy General Tools & Instruments Rain Gauges Items Including:
General Digital Rain Guage and Remote Rain Collector with MIN/MAX and.
More Details
Nalgene Specialty Dispensers
Buy Nalgene Specialty Dispensers Items Including:
Thermo Scientific Nalgene Filling bell for liquid transfer, 4-1/2"H x.
More Details
Specialty Dispensers
Buy Specialty Dispensers Items Including:
Thermo Scientific Nalgene Filling bell for liquid transfer, 4-1/2"H x.
More Details
Digital Traceable Ultra Low Freezer Thermometer
Buy Digital Traceable Ultra Low Freezer Thermometer Items Including:
FRIO-TEMP ULTRA LOW FREEZER THERMOMETER, -130/80F 145MM LENGTH, BOTTLE
Frio-temp Certified Verification Thermometer For Ultra Low Freezers, P
Frio-temp Precision Verification Thermometer For Ultra Low Freezers, P.
More Details
Low-Flow Piston Pump System
Buy Low-Flow Piston Pump System Items Including:
Piston Metering Pump, 3/32" stainless head, 0.002 to 2.5 mL/min range, (1LMP)
Piston Metering Pump, 1/8" stainless head, 0.003 to 5 mL/min range, 60 (1SMP)
Piston Metering Pump, 3/32" stainless head, 0.003 to 5 mL/min range, 6 (2LMP).
More Details
Cole-Parmer Single Glass and Quartz Cuvettes
Buy Cole-Parmer Single Glass and Quartz Cuvettes Items Including:
Cole-Parmer® Fluorimeter cuvettes with cover of PTFE resin; glass; (YO-83303-02)
Cole-Parmer® Fluorimeter cuvettes with cover of PTFE resin; glass; (YO-83303-03)
Cole-Parmer® Fluorimeter cuvettes with cover of PTFE resin; glass; (YO-83303-04).
More Details
Colored Glass Vials with Dropper
Buy Colored Glass Vials with Dropper Items Including:
E-C Vial RBR w/o cap Clear 2ml (200 cs) (224701)
E-C Vial RBR w/o cap Clear 4ml (200 cs) (224702)
E-C Vial RBR w/o cap Clear 8ml (200 cs) (224704)
E-C Vial RBR w/o cap Clear 12ml (200 cs) (224705)
E-C Vial RBR w/o cap Clear 16ml (200 cs) (224706).
More Details
Refractometer Fluid
Buy Refractometer Fluid Items Including:
Reichert Industrial Fluid Refractometer, 0 to 10 scale (1.3330 to 1.34
Reichert Industrial Fluid Refractometer, 0 to 30 scale (1.3330 to 1.38
Reichert Coolant/Battery Acid Refractometer; Ethylene Glycol: -60 to 3.
More Details
Self Contained Eyewash Station
Buy Self Contained Eyewash Station Items Including:
Encon Aquarion Eyewash Station
Honeywell100% Sterile Primary Eyewash Station with Alarm
Portable Pressurized Eyewash with Drench Hose, 10 gallon
Portable Pressurized Eyewash with Drench Hose, 15 gallon
Bradley On-Site Portable Gravity-Fed Eyewash.
More Details
Weatherlink Weather Stations
Buy Weatherlink Weather Stations Items Including:
Davis Instruments WeatherLink Software for Vantage Pro 2, Mac
Davis Instruments WeatherLink Software (with data logging feature) for (6510SER).
More Details
70618
results
<<<Previous
896
897
898
899
900
901
902
903
904
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
100
200
300
400
500
600
700
800
810
820
830
840
850
860
870
880
885
886
887
888
889
890
891
892
893
894
895
896
897
898
899
900
901
902
903
904
905
906
907
908
909
910
911
912
913
914
920
930
940
950
960
970
980
990
1,000
1,100
1,200
1,300
1,400
1,500
1,600
1,700
1,800
2,000
2,354
Bel-a Safety Labeled Wide-Mouth Wash Bottles to Weatherlink Weather Stations
>
31125329-s-toc_2 31125329stoc2