Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 1000

Humidity Control Co2 Incubator   to   Humidity Levels

Image
  Product Detail
More Detail

Humidity Control Co2 Incubator

Buy Humidity Control Co2 Incubator Items Including:
  • 1000-liter Plant Growth Chamber With Humidity Control, 230VAC (AAHA1011) (YO-39353-20)
  • 1000-liter Plant Growth Chamber With Humidity Control, 230VAC (AAHA1011) (YO-39353-25)
  • GE (Telaire) Dual Position CO2, Temperature, and Humidity M.

Humidity Control Equipment

Buy Humidity Control Equipment Items Including:
  • Digital Humidity Controller (5100-240)
  • 1000-liter Plant Growth Chamber With Humidity Control, 230VAC (AAHA1011) (YO-39353-20)
  • Controlled Humidity Chamber, 115 VAC (5506-115V)
  • AC powered nanodac with two logic, one relay output and the humidity b.

Humidity Control System

Buy Humidity Control System Items Including:
  • WSC-5-DDO : Wind Speed Controller Dual Set Point
  • WSC-5-SDOR : Wind Speed Controller Single Set Pt.
  • LabMaster Standard Water Activity Instrument, with Full Temperature Co
  • LabMaster AdvancedWater Activity Instrument, with Full Temperature Con.

Humidity Controller Electronic

Buy Humidity Controller Electronic Items Including:
  • WSC-5-SDOR : Wind Speed Controller Single Set Pt.
  • WSC-5-DDO : Wind Speed Controller Dual Set Point.

Humidity Controller for Display

Buy Humidity Controller for Display Items Including:
  • 1/32-DIN Temp
  • controller with single-line display, SSRD, relay, 100- (330000000)
  • 1/32-DIN Temp
  • controller with single-line display, relay, relay, 100 (331100000)
  • 1/32-DIN controller with single-line display, RS-232, SSRD, relay, 100 (330 000 200).

Humidity Controller Hum 1

Buy Humidity Controller Hum 1 Items Including:
  • Temperature Controller, RTD, 1/16-DIN, Relay Output, 1 alarm (DC1014CR-101-000-E)
  • Temperature Controller, RTD, 1/16-DIN, Current Output, 1 alarms (DC1014CR-301-000-E)
  • Temperature Controller, TC, 1/4-DIN, Relay Output, 1 alarms, (DC1044CT-101-000-E).

Humidity Controller Switch

Buy Humidity Controller Switch Items Including:
  • Temperature Switches: temperature trip point; 100F (AD148-12/100F)
  • Configuration Key for Love Controllers
  • Thermocouple Temperature Controller, Type K and J, 110V, °F (TCS-4010)
  • Thermocouple Temperature Controller, Type K and J, 110V, °C (TCS-4011).

Humidity Controller with Multiple Input Points

Buy Humidity Controller with Multiple Input Points Items Including:
  • Digi-Sense 1-Zone benchtop dual display Limit control, type J input, a (YO-36225-76)
  • 1/16 DIN controllers with universal input and fuzzy logic, two SPST re (16A2133)
  • 1/16 DIN controllers with universal input and fuzzy logic, two SSR and (16A2111).

Humidity Controller with Probe

Buy Humidity Controller with Probe Items Including:
  • Cole-Parmer 100 ohm Platinum RTD Probes; 5"L
  • Cole-Parmer 100 ohm Platinum RTD Probes; 9"L
  • Type T High-temperature Thermocouple probes; 12"L (050797B-T)
  • Humidity Probe, 0-3V Output (THC-P)
  • Thermistor Probe, 1000 ohm, 5' , PVC (TS-5).

Humidity Controller with Remote Sensor

Buy Humidity Controller with Remote Sensor Items Including:
  • Cole-Parmer Thermohygrometer with remote sensor
  • Wireless Thermometer Set; Includes Monitor And One Remote Sensor (4115CP)
  • Wireless Thermometer Remote Sensor Module for p/n 94460-78
  • Wireless Thermometer Remote Bottle Sensor Module for p/n 37804-09.

Humidity Data

Buy Humidity Data Items Including:
  • Track It™ temperature and humidity data logger with display and sta (YO-30003-02)
  • LASCAR EL-WiFi-TH Wi-Fi Temperature & Humidity Data Logging Sensor
  • MicroLogPROII Temperature/Humidity Data Logger EC850
  • Temperature/Humidity Strip Chart Recorder.

Humidity Data Loggers

Buy Humidity Data Loggers Items Including:
  • USB Temperature/Humidity Data Logger, 16,000+ readings, 1 year battery
  • MicroLogPROII Temperature/Humidity Data Logger EC850
  • Track It™ temperature data logger with display and standard battery
  • Track It™ data logger USB interface cable (YO-30003-04).

Humidity Data Loggers and Recorders

Buy Humidity Data Loggers and Recorders Items Including:
  • USB Temperature/Humidity Data Logger, 16,000+ readings, 1 year battery
  • MicroLogPROII Temperature/Humidity Data Logger EC850
  • Track It™ temperature data logger with display and standard battery
  • Track It™ data logger USB interface cable (YO-30003-04).

Humidity Data Logging

Buy Humidity Data Logging Items Including:
  • Davis Instruments WeatherLink Software (with data logging feature) for (6510SER)
  • Humidity-Temperature Meter w/ SD Card Data Logging
  • THERM-A-LERT Temperature Data Logger with Wireless Transceiver
  • THERM-A-LERT Wireless Data Logger with precision RTD.

Humidity Dataloggers

Buy Humidity Dataloggers Items Including:
  • Extech Compact temperature and humidity datalogger kit
  • Jumbo LED display temperature/humidity datalogger (LTH8809DL)
  • Digital Temperature/Humidity Datalogger (RHTEMP2000)
  • Data View Datalogger, Temperature/Humidity Datalogger with Graphic Dis (DVTH).

Humidity Detection

Buy Humidity Detection Items Including:
  • General Deluxe Lightning/Storm Detector LD50.

Humidity Detectors

Buy Humidity Detectors Items Including:
  • General Personal Lightening Detector LD7
  • Grain moisture detector.

Humidity Dew

Buy Humidity Dew Items Including:
  • GE (General Eastern) Industrial relative Humidity/Dew Point Analyzer, (MR2350) (YO-37405-04)
  • Rh / Dew Point And Temperature Transmitter (display - Remote Probe) (DMT340-8V0B1A1BEA1B001A1ACB0A0)
  • Industrial dew Point Transmitter with Plastic Housing.

Humidity Dew Point Chart

Buy Humidity Dew Point Chart Items Including:
  • Rh / Dew Point And Temperature Transmitter (no Display - Remote Probe) (DMT340-8V0B0A1BEA1B001A1ACB0A0)
  • GE (General Eastern) Industrial Dew Point Transmitter with Display
  • Industrial dew Point Transmitter with Plastic Housing
  • Dew Point Meter (DPMi).

Humidity Dial

Buy Humidity Dial Items Including:
  • Oakton Large-Dial Thermohygrometer
  • 3" Dial, Silicone-filled Bottom-connected Dial Dual-Scale Industrial B (31090T006A4SF)
  • 3" Dial, Silicone-filled Bottom-connected Dial Dual-Scale Industrial B (31120T006A4SF).

Humidity Display

Buy Humidity Display Items Including:
  • Dickson 8" Temperature/humidity Recorder With Digital Display (TH802) (YO-80001-52)
  • Dickson 8" Temperature/humidity Recorder With Probe, Dig
  • Display And (TH803) (YO-80001-54)
  • Dickson 6" Temperature/Humidity Recorder with LCD Display.

Humidity Gauges

Buy Humidity Gauges Items Including:
  • Industrial Thermocouple Element, 12"L, 20 Gauge, K type (5504A-K-20-12-0)
  • Industrial Thermocouple Element, 12"L, 20 Gauge, J type (5504A-J-20-12-0)
  • Rainwise Dual Display Rain Gauge, 30ft cable
  • Rainwise Wireless Dual Display Rain Gauge.

Humidity Generator

Buy Humidity Generator Items Including:
  • 25 Pack Of Thermal Pads (Used With Heatsinks to dissipate generated he.

Humidity Indicator Card

Buy Humidity Indicator Card Items Including:
  • Cole-Parmer Digital Thermohygrometer with Dew Point and Memory Card.

Humidity Indicator Strips

Buy Humidity Indicator Strips Items Including:
  • Seven-Point Horizontal Thermometer with Adhesive Backing, -22 to 32de (YO-90308-20)
  • Temperature Labels, Refrigerator (YO-90308-80)
  • Fortyeight-point vertical thermometer with Magnetic Backing, 26 to 120 (YO-08078-48).

Humidity Indicators

Buy Humidity Indicators Items Including:
  • NIST-Traceable Humidity Card (4192CP)
  • Calibration kit for Extech humidity indicators models 37809-16 and 378
  • Cole-Parmer Humidity/Thermometer Meter
  • Cole-Parmer Brass Case Indicator, % RH and °F/°C
  • Large digital clock with humidity and temperature.

Humidity Indicators (Digital)

Buy Humidity Indicators (Digital) Items Including:
  • Digital Thermohygrometer
  • Remote temperature/humidity monitor with alarm
  • Digital Termperature and Humidity Monitor
  • Cole-Parmer Digital Thermohygrometer with Dew Point and Memory Card
  • Cole-Parmer Digital Thermohygrometer with Dew Point and Wet-bulb.

Humidity Instruments

Buy Humidity Instruments Items Including:
  • Davis Instruments Mark 3 Sextant
  • Davis Instruments Mark 15 Sextant
  • Davis Instruments Mark 25 Sextant
  • Davis Instruments Vantage Pro2 Plus Cabled Weather Station System
  • Davis Instruments AC-powered Repeater

Humidity Labels

Buy Humidity Labels Items Including:
  • Cole-Parmer Psychrometer, F (22014 ColeParmer labeled)
  • HOT Hand Reversible Indicating Label, Changes from orange to yellow at (4071COPADP10)
  • HOT Hand Reversible Indicating Label, Changes from Red to black at 157 (4077COPADP10).

Humidity Levels

Buy Humidity Levels Items Including:
  • Fluke TiS Thermal Imager - Entry Level (TiS)
  • Fluke Ti100 Thermal Imager - Entry Level
  • Flir i5 Compact Thermal Imager (100 x 100)
  • Extech Technician's Kit with i5 Thermal Inager
  • Extech TM300 Dual input Type K/J thermometer with Differential/Min/Max.



Humidity Control Co2 Incubator to Humidity Levels

>