Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 12
Accessories for Positive Displacement Pumps
to
Accident Prevention Tags Record Tags
Image
Product Detail
More Detail
Accessories for Positive Displacement Pumps
Buy Accessories for Positive Displacement Pumps Items Including:
Liquid level controller, 1/4" to 1" pumps, Normally closed, Close, PVC
Liquid level controller, 1-1/2" to 3" pumps, Normally closed, Close, S
Pump, AODD Service Kit for SKU 75320-91
Pump, AODD Service Kit for SKU 75320-93.
More Details
Accessories for Reichert Abbe Refractometers
Buy Accessories for Reichert Abbe Refractometers Items Including:
Replacement standard solution, 7 mL of 1-bromonaphthalene.
More Details
Accessories for Respirator Cleaning Kit
Buy Accessories for Respirator Cleaning Kit Items Including:
Respirator Cleaning Kit
Replacement Soap Packets for Respirator Cleaning Kit, 90/pack
Replacement Storage Bags for Respirator Cleaning Kit, 100/pack.
More Details
Accessories for Retsch Air-Jet Sieving Machine
Buy Accessories for Retsch Air-Jet Sieving Machine Items Including:
Filter Bags, 5/pack
HEPA Filter (YO-59996-33)
Automatic Vacuum Regulator
Replacement Cover
Industrial Vacuum Cleaner.
More Details
Accessories for Salvis Incucenter Programmable Incubators
Buy Accessories for Salvis Incucenter Programmable Incubators Items Including:
Extra Shelf for 2.8 cu ft Incucenter Incubator (YO-39701-52)
Extra Shelf for 5.6 cu ft Incucenter Incubator (YO-39701-54)
Extra Shelf for 8.5 cu ft Incucenter Incubator (YO-39701-56)
Extra Shelf for 14.0 cu ft Incucenter Incubator (YO-39701-58).
More Details
Accessories for Shakers
Buy Accessories for Shakers Items Including:
Universal 18" x 18" shaker platform
Universal 18" x 24" shaker platform
Universal platform, 14" x 14" for Lab
Companion Dual-Action Shaker
Universal platform, 18" x 18" for Lab
More Details
Accessories for Sieves
Buy Accessories for Sieves Items Including:
W.S
Tyler Sound Enclosure for Ro-Tap II Sieve Shaker, 34"W x 31"H x 3
3" Diameter Stainless Steel Full Height Sieve Pan
Round Sieve Brush for 120 Mesh Sieves and Finer, Nylon Bristles
Cover with Porthole for Vibratory Sieve Shaker.
More Details
Accessories for Sludge Judge Sampling Systems
Buy Accessories for Sludge Judge Sampling Systems Items Including:
Sludge Judge II, replacement bottom section with valve
Sludge Judge II Replacement Middle/Extension Section
Sludge Judge II Replacement Top Section with Rope
Sludge Judge Replacement Middle/Extension Section
Sludge Judge Sampler Canvas Sorage Bag.
More Details
Accessories for Smart Gaskets
Buy Accessories for Smart Gaskets Items Including:
Probe,type K,short,w/3ft Cable For Smart Gasket System (YO-30545-90)
T/c Wire Kapton, 10ft,24 Gauge For Smart Gasket System (YO-30545-93)
Hole plug for Smart Gasket System (YO-30545-98)
Sampling tube for Smart Gasket System (YO-30545-99).
More Details
Accessories for Sonicator Ultrasonic Processors
Buy Accessories for Sonicator Ultrasonic Processors Items Including:
Open end wrench (requires 2) for use with ultrasonic processors
Replacement tube holder (20 tubes) for use with ultrasonic processors
Dual Horn (without probes) for use with ultrasonic processors
Stand with Clamp for use with ultrasonic processors.
More Details
Accessories for Spectroscopy
Buy Accessories for Spectroscopy Items Including:
Cole-Parmer Spectroscopy Standard Kit 1, includes 4 pcs, for UV-VIS Ph
GENESYS 10 nanoCell accessory, 0.2mm pathlength (222-227700)
GENESYS 10 nanoCell accessory, 1.0mm pathlength (222-227800)
Thermo Scientific NanoDrop Lite Printer Accessory.
More Details
Accessories for Standard Ultrasonic Cleaners
Buy Accessories for Standard Ultrasonic Cleaners Items Including:
Solid tray for Cole-Parmer 1/2 Gallon and Stainless Steel 3/4 Gallon C
Perforated tray for Cole-Parmer 1/2 Gallon and Stainless Steel 3/4 Gal
Mesh tray for Cole-Parmer 1/2 Gallon and Stainless Steel 3/4 Gallon Cl.
More Details
Accessories for Stroboscopes
Buy Accessories for Stroboscopes Items Including:
Spare For Pk2 Stroboscope Only
Charger for BB 115 Battery Powered Stroboscope (YO-98806-83)
Replacement strobe lamp for all Cole-Parmer digital stroboscopes (YO-87000-82)
Latched carrying case for all Cole-Parmer digital stroboscopes (YO-87000-90).
More Details
Accessories for Tbs Shur/Wave Laboratory Microwave
Buy Accessories for Tbs Shur/Wave Laboratory Microwave Items Including:
Tube adapter fitting for SHUR/Wave Laboratory Microwave
SHUR/Wave™ Temperature Probe Insulators, 25cm length; 2ea/pk
Small Rack Assembly with glass bowl, 36 cassette capacity for SHUR/Wav
Medium rack assembly with glass bowl, 64 cassett capacity.
More Details
Accessories for Viscosity Baths
Buy Accessories for Viscosity Baths Items Including:
Bath oil, 6 gallons (9726-L20)
Diaphragm vacuum pump, 120 VAC (9726-L96)
Neoprene viscometer holder for all Cannon-Fenske types
Plastic viscometer holder for Cannon-Ubbelohde viscometers
Neoprene viscometer holder for Cannon-Ubbelohde viscometers.
More Details
Accessories for Waring Four-Liter Blenders
Buy Accessories for Waring Four-Liter Blenders Items Including:
Cool base container for four-liters containers (2610C)
Repl Container 4 Liter SS (CAC70)
Repl Blade Assembly 4L (010698).
More Details
Accessories for Waring One-Liter Blenders
Buy Accessories for Waring One-Liter Blenders Items Including:
Mini containers with screw SS lids for one-liter blenders; 3 to 20 mL (8575)
Container SS 1-liter (SS515)
Replacement Blade Assembly for 1 Liter Waring Blender (003548)
Stainless steel container, 1 liter for Hazard Location Motor Base; ite (HL515).
More Details
Accessories for Waring Two-Liter Blenders
Buy Accessories for Waring Two-Liter Blenders Items Including:
Replacement Blade Assembly F/2l.
More Details
Accessories for Waterless Ice Buckets
Buy Accessories for Waterless Ice Buckets Items Including:
Chill packs for waterless ice bucket
Waterless ice bag kit; 2L beads, two chill packs
Bead bag for waterless ice baths or buckets.
More Details
Accessories Nikon
Buy Accessories Nikon Items Including:
10:1 Stereozoom Microscope 35-mm Camera Adapter, Nikon (T2-5).
More Details
Accessories Water
Buy Accessories Water Items Including:
Global Water Turbidity Sensor (WQ730)
pH Con Accessory Kit (YO-35630-69)
BOD Accessory Kit for Hach Dissolved Oxygen Meters
Ultrapure water DO connection kit, for use with PVC DO electrode
Oakton deionized water rinse solution, box of 20 pouches..
More Details
Accessory Kit
Buy Accessory Kit Items Including:
pH Con Accessory Kit (YO-35630-69)
BOD Accessory Kit for Hach Dissolved Oxygen Meters
Oakton Dissolved Oxygen Meter Accessory Kit
Extech 479097 Accessory Kit for 475040/475044 Force Gauges
2 each-hoo
More Details
Accessory Tips for Mla Pipettes
Buy Accessory Tips for Mla Pipettes Items Including:
Stacked Rack Of Pipette Tips For Use With mLa™ Pipettes, 201-1000mL
Macro Graduated Pipette Tips For Use With mLa™ Pipettes, 1-5mL, 100
Ovation & mLa™ Graduated Pipette Tips For Use With mLa™ Pipettes.
More Details
Accessory Top Surface
Buy Accessory Top Surface Items Including:
Work Surface - Metallic Tray & Tempered Glass Top For 33520-20 And -22 (YO-33520-27)
Work Surface - Metallic Tray & Tempered Glass Top For 33520-30 And -32 (YO-33520-92).
More Details
Accident Prevention
Buy Accident Prevention Items Including:
Tag, Danger Defective Equipment, Back A, 5 7/8" X 3 1/8", PF-Cardstock (MDT122CTP)
Tag, Danger Defective Equipment, Back B, 5 7/8" X 3 1/8", PF-Cardstock (MDT229CTP).
More Details
Accident Prevention Signs
Buy Accident Prevention Signs Items Including:
Tag, Danger Defective Equipment, Back A, 5 7/8" X 3 1/8", PF-Cardstock (MDT122CTP)
Tag, Danger Defective Equipment, Back B, 5 7/8" X 3 1/8", PF-Cardstock (MDT229CTP).
More Details
Accident Prevention Signs and Tags
Buy Accident Prevention Signs and Tags Items Including:
Tag, Danger Do Not Use, Back A, 5 7/8" X 3 1/8", PF-Cardstock (MDT106CTP)
Tag, Danger Do Not Use, Back A, 5 7/8" X 3 3/8", RV-Plastic (MDT106PTP)
Tag, Danger Do Not Use, Back B, 5 7/8" X 3 3/8", RV-Plastic (MDT217PTP).
More Details
Accident Prevention Tags
Buy Accident Prevention Tags Items Including:
Tag, Danger Defective Equipment, Back A, 5 7/8" X 3 1/8", PF-Cardstock (MDT122CTP)
Tag, Caution Unsafe Do Not Use, Back A, 5 7/8" X 3 1/8", PF-Cardstock (MCGT225CTP).
More Details
Accident Prevention Tags Danger Defective Equipment
Buy Accident Prevention Tags Danger Defective Equipment Items Including:
Tag, Danger Defective Equipment, Back A, 5 7/8" X 3 1/8", PF-Cardstock (MDT122CTP)
Tag, Danger Defective Equipment, Back B, 5 7/8" X 3 1/8", PF-Cardstock (MDT229CTP).
More Details
Accident Prevention Tags Record Tags
Buy Accident Prevention Tags Record Tags Items Including:
Tag, Emergency Shower & Eye Wash Test Record, 5 7/8" X 3 1/8", PF-Card (MGT207CTP)
Tag, Forklift Inspection, 5 7/8" X 3 3/8", RV-Plastic (TRS305PTP)
Tag, Safety Inspection, 5 7/8" X 3 1/8", PF-Cardstock (TRS315CTP).
More Details
70618
results
<<<Previous
8
9
10
11
12
13
14
15
16
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Accessories for Positive Displacement Pumps to Accident Prevention Tags Record Tags
>
31125329-s-toc_2 31125329stoc2