Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 2

A&D Engineering Weighing Balance Accessories   to   a&D Sk Stainless Steel Washdown Scales

Image
  Product Detail
More Detail

A&D Engineering Weighing Balance Accessories

Buy A&D Engineering Weighing Balance Accessories Items Including:
  • USB Interface for A&D Weighing Newton Balance (EJ-02)
  • Replacement printer paper for A&D Weighing Printer.

A&D Everest Eki Portable Balances

Buy A&D Everest Eki Portable Balances Items Including:
  • A&D Eki Portable Balance, 120 g x 0.01 g
  • A&D Eki Portable Balance, 1200 g capacity
  • A&D Eki Portable Balance, 2000 g capacity
  • A&D Eki Portable Balance, 3000 g capacity
  • A&D Eki Portable Balance, 12000 g capacity.

A&D Fc-Si and Fc-I Counting Scales

Buy A&D Fc-Si and Fc-I Counting Scales Items Including:
  • A&D FC-Si Counting Scale, 500g capacity
  • A&D FC-i Counting Scale, 5000g capacity
  • A&D FC-i Counting Scale, 10,000g capacity
  • A&D FC-i Counting Scale, 20,000g capacity
  • A&D FC-i Counting Scale, 50,000g capacity.

A&D Fg-K Industrial Scales

Buy A&D Fg-K Industrial Scales Items Including:
  • A&D FG-K Industrial Scale, 60 lb x 0.01 lb with display column
  • A&D FG-K Industrial Scale, 150 lb x 0.02 lb with display column.

A&D Fsi Washdown Industrial Scales

Buy A&D Fsi Washdown Industrial Scales Items Including:
  • A&D FSi Washdown Industrial Scale, 6 kg (15 lb)
  • A&D FSi Washdown Industrial Scale, 15 kg (30 lb).

A&D Fx-I Toploading Balances

Buy A&D Fx-I Toploading Balances Items Including:
  • A&D FX-i Toploading Balance 122g X 0.001g Ext.Calibration, Comparator,
  • A&D FX-iWP Toploading Balance, 122g X 0.001g Ext.Calibration, Comparat (FX-120IWP)
  • A&D FX-iWP Toploading Balance, 220g X 0.001g Ext.Calibration, Comparat (FX-200IWP).

A&D Fx-Iwp Waterproof Toploading Balances

Buy A&D Fx-Iwp Waterproof Toploading Balances Items Including:
  • A&D FX-iWP Toploading Balance, 1220g X 0.01g Ext.Calibration, Comparat (FX-1200IWP+230VAC)
  • A&D FX-iWP Toploading Balance, 2200g X 0.01g Ext.Calibration, Comparat (FX-2000IWP+230VAC).

A&D Galaxy Analytical Balances

Buy A&D Galaxy Analytical Balances Items Including:
  • A&D Galaxy HR-AZ Analytical Balance, 102g x 0.1mgwith Internal Calibra
  • A&D Galaxy HR-AZ Analytical Balance, 152g x 0.1mg with Internal Calibr
  • A&D Galaxy HR-AZ Analytical Balance, 252g x 0.1mg with Internal Calibr.

A&D Gemini Autocalibrating Analytical Balances

Buy A&D Gemini Autocalibrating Analytical Balances Items Including:
  • A&D Gemini Autocalibrating Analytical Balance, 120 G
  • A&D Gemini Autocalibrating Analytical Balance, 210 G.

A&D Gf Toploading Balances

Buy A&D Gf Toploading Balances Items Including:
  • A&D GF Toploading Balance, 410g x 0.001g External Calibration, RS232,
  • A&D GF Toploading Balance, 610g x 0.001g External Calibration, RS232,
  • A&D GF Toploading Balance, 810g x 0.001g External Calibration, RS232,.

A&D Gp Washdown Industrial Scales

Buy A&D Gp Washdown Industrial Scales Items Including:
  • A&D GP Washdown Industrial Scale, 31 kg x 0.1 g 115VAC
  • A&D GP Washdown Industrial Scale, 100 kg x 1 g, 115VAC
  • A&D GP Washdown Industrial Scale, 12kg x 0.1 g
  • A&D GP Washdown Industrial Scale, 21kg x 0.1 g.

A&D Gx Toploading Balances

Buy A&D Gx Toploading Balances Items Including:
  • A&D GX Toploading Balance, 210 g x 0.001 g Internal Calibration
  • A&D GX Toploading Balance, 410 g x 0.001 g Internal Calibration
  • A&D GX Toploading Balance, 810g x 0.001g Internal Calibration, RS232,.

A&D Hc-I Counting Scales

Buy A&D Hc-I Counting Scales Items Including:
  • A&D HC-i Counting Scale, 3 kg/6 lb
  • A&D HC-i Counting Scale, 15 lb x 0.002 lb
  • A&D HC-i Counting Scale, 30 lb x 0.005 lb
  • A&D HC-i Counting Scale, 60 x 0.01 lb.

A&D High Capacity Scales

Buy A&D High Capacity Scales Items Including:
  • Ohaus Explorer High Capacity Toploading Balance, 12,000g External Cali
  • Ohaus Explorer High Capacity Toploading Balance, 22,000g External Cali
  • Ohaus Explorer High Capacity Toploading Balance, 32,000g External Cali.

A&D Hlwp Portable Washdown Balances

Buy A&D Hlwp Portable Washdown Balances Items Including:
  • A&D HLWP Portion Control Washdown Scale, 300 G X 0.1 G
  • A&D HLWP Portion Control Washdown Scale, 1000 G X 0.5 G
  • A&D HLWP Portion Control Washdown Scale, 3000 G X 1 G
  • A&D HLWP Portion Control Washdown Scale, 3000 G X 1 G (ntep).

A&D Hlwp Portion Control Washdown Scales

Buy A&D Hlwp Portion Control Washdown Scales Items Including:
  • A&D HLWP Portion Control Washdown Scale, 300 G X 0.1 G
  • A&D HLWP Portion Control Washdown Scale, 1000 G X 0.5 G
  • A&D HLWP Portion Control Washdown Scale, 3000 G X 1 G
  • A&D HLWP Portion Control Washdown Scale, 3000 G X 1 G (ntep).

A&D Ht Portable Scales

Buy A&D Ht Portable Scales Items Including:
  • A&D HT-Series Compact Scales 310g x 0.1g incl carry case
  • A&D HT-Series Compact Scales 510g x 0.1g incl carry case
  • A&D HT-Series Compact Scales 3100g x 1g incl carry case
  • A&D HT-Series Compact Scales 5100g x 1g incl carry case.

A&D Hw Industrial Scales

Buy A&D Hw Industrial Scales Items Including:
  • A&D HW Industrial Scale, 100 kg x 0.01 kg (Triple Range)
  • A&D HW Industrial Scale, 3/6/15 kg x 0.001/0.002/0.005 kg (Triple Rang
  • A&D HW Washdown Industrial Scale, Capacity 150lb.
  • A&D HW Washdown Industrial Scale, Capacity 200lbs..

A&D Hw Washdown Industrial Scales

Buy A&D Hw Washdown Industrial Scales Items Including:
  • A&D HW Washdown Industrial Scale, Capacity 150lb.
  • A&D HW Washdown Industrial Scale, Capacity 200lbs.
  • A&D HW Washdown Industrial Scale, 3/6/15 kg x 0.001/0.002/0.005 g (Tri
  • A&D HW Washdown Industrial Scale, Capacity 132/300/500lbs..

A&D Moisture Determination Balances

Buy A&D Moisture Determination Balances Items Including:
  • A&D Moisture Determination Balance, ML-50, 51 g x 0.005 g, 115 VAC
  • A&D Moisture Determination Balance, MF-50, 51 g x 0.002 g, 115 VAC
  • A&D Moisture Determination Balance, MX-50, 51 g x 0.001 g, 115 VAC.

A&D Newton Ej Portable Balances

Buy A&D Newton Ej Portable Balances Items Including:
  • A&D Newton EJ Portable Balance, 120 G X 0.01 G; 115VAC
  • A&D Newton EJ Portable Balance, 210 G X 0.01 G; 115VAC
  • A&D Newton EJ Portable Balance, 310 G X 0.01 G; 115VAC
  • A&D Newton EJ Portable Balance, 410 G X 0.01 G; 115VAC.

A&D Ntep-Approved Balances

Buy A&D Ntep-Approved Balances Items Including:
  • A&D NTEP-Approved Balance, 5,000 g
  • Ohaus Adventurer Pro Precision Balance AV53N, 51gx0.02gNTEP Approved,W
  • Ohaus Adventurer Pro Precision Balance AV212N, 210g x 0.1g, NTEP App
  • Ohaus Adventurer Pro Precision Balance AV412N 410g x 0.1g, NTEP Appr.

A&D Orion Analytical Balances

Buy A&D Orion Analytical Balances Items Including:
  • A&D Orion Analytical Balance, 60 g
  • A&D Orion Analytical Balance, 120 g
  • A&D Orion Analytical Balance, 210 g
  • A&D Orion Analytical Balance, 310 g x 0.1 mg, 120 VAC.

A&D Portable Toploading Balances

Buy A&D Portable Toploading Balances Items Including:
  • Symmetry EC II Portable Toploading Balance, 120g X 0.005g (220 VAC) (YO-20000-27)
  • Symmetry EC II Portable Toploading Balance, 400g X 0.01g (220 VAC) (YO-20000-28)
  • Symmetry EC II Portable Toploading Balance, 2000g X 0.05g (220 VAC) (YO-20000-29).

A&D Pv Handheld Balances

Buy A&D Pv Handheld Balances Items Including:
  • A&D PV Handheld Balance, 60 g x 0.01 g
  • A&D PV Handheld Balance, 100 g x 0.1 g
  • A&D PV Handheld Balance, 200 g x 0.1 g.

A&D Se Washdown Industrial Scales

Buy A&D Se Washdown Industrial Scales Items Including:
  • A&D SE Washdown Industrial Scale, 66lb x 0.01lb/30kg x 0.005kg IP68, B
  • A&D SE Washdown Industrial Scale, 130lb x 0.02lb/60kg x 0.01kg IP68, B (SE-60KAM)
  • A&D SE Washdown Industrial Scale, 330lb x 0.05lb/150kg x 0.02kg IP68, (SE-150KAM).

A&D Semi/Micro Analytical Balances

Buy A&D Semi/Micro Analytical Balances Items Including:
  • Sartorius Cubis Semi-Micro Balance, 220 G X 0.01 Mg, 120VAC (MSA225S-000-DU)
  • Sartorius Cubis Analytical Balance, 1,200 G X 1 MG, 120VAC (MSA1203S-000-DE)
  • Sartorius Cubis Analytical Balance, 2,200 G X 1 MG, 120VAC (MSA2203S-000-DE).

A&D Sj Portion Scales

Buy A&D Sj Portion Scales Items Including:
  • A&D SJ Series Digital Portion Scale, 1000 G X 0.5 G
  • A&D SJ Series Digital Portion Scale, 2000 G X 1 G
  • A&D SJ Series Digital Portion Scale, 12 Kg X 0.005 Kg
  • A&D SJ Series Digital Portion Scale, 20 Kg X 0.01 Kg.

A&D Sj Series Digital Portion Scales

Buy A&D Sj Series Digital Portion Scales Items Including:
  • A&D SJ Series Digital Portion Scale, 1000 G X 0.5 G
  • A&D SJ Series Digital Portion Scale, 2000 G X 1 G
  • A&D SJ Series Digital Portion Scale, 12 Kg X 0.005 Kg
  • A&D SJ Series Digital Portion Scale, 20 Kg X 0.01 Kg.

A&D Sk Stainless Steel Washdown Scales

Buy A&D Sk Stainless Steel Washdown Scales Items Including:
  • A&D Portable Stainless Steel Washdown Scale, 1000 g (SK-1000WP)
  • A&D Portable Stainless Steel Washdown Scale, 2000 g (SK-2000WP)
  • A&D Portable Stainless Steel Washdown Scale, 5000 g (SK-5000WP).

70618 results


A&D Engineering Weighing Balance Accessories to a&D Sk Stainless Steel Washdown Scales

>