Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 20
Accumetric
to
Acetal Check Valves
Image
Product Detail
More Detail
Accumetric, Llc Bath & Circulator Accessories
Buy Accumetric, Llc Bath & Circulator Accessories Items Including:
Bath Fluid, Silicone 710 (hot) (YO-01294-00)
Bath Fluid, Silicone 200 20 (hot) (YO-01294-40)
Bath Fluid, Silicone 210 (YO-01294-60)
Bath Fluid, Dynalene HC-50.
More Details
Accumetric, Llc Bath Fluids
Buy Accumetric, Llc Bath Fluids Items Including:
Bath Fluid, Silicone 710 (hot) (YO-01294-00)
Bath Fluid, Silicone 200 20 (hot) (YO-01294-40)
Bath Fluid, Silicone 210 (YO-01294-60)
Bath Fluid, Dynalene HC-50.
More Details
Accumetric, Llc Baths & Circulators
Buy Accumetric, Llc Baths & Circulators Items Including:
Bath Fluid, Silicone 710 (hot) (YO-01294-00)
Bath Fluid, Silicone 200 20 (hot) (YO-01294-40)
Bath Fluid, Silicone 210 (YO-01294-60)
Bath Fluid, Dynalene HC-50.
More Details
Accumetric, Llc Lab Equipment
Buy Accumetric, Llc Lab Equipment Items Including:
Bath Fluid, Silicone 710 (hot) (YO-01294-00)
Bath Fluid, Silicone 200 20 (hot) (YO-01294-40)
Bath Fluid, Silicone 210 (YO-01294-60)
Bath Fluid, Dynalene HC-50.
More Details
Accupenser Jr Bottle Top Dispenser
Buy Accupenser Jr Bottle Top Dispenser Items Including:
Cole-Parmer Bottletop Dispenser .25-2.5 mL
Cole-Parmer Bottletop Dispenser .25-2.5 mL with 100 mL Glass Bottle
Cole-Parmer Bottletop Dispenser 2.5-30 mL with 500 mL Glass Bottle
Cole-Parmer Bottletop Dispenser 5-60 mL with 1000 mL Glass Bottle.
More Details
Accuracy of Cannon Fenske Routine Viscometer
Buy Accuracy of Cannon Fenske Routine Viscometer Items Including:
Cannon Cannon-Fenske routine viscometer, size 25
Cannon Cannon-Fenske routine viscometer, size 50
Cannon Cannon-Fenske routine viscometer, size 75
Cannon Cannon-Fenske routine viscometer, size 100
Cannon Cannon-Fenske routine viscometer, size 150.
More Details
Accuracy of in Tire Pressure Transducer
Buy Accuracy of in Tire Pressure Transducer Items Including:
AccuSense™ Pressure Transducer, 30-0" HG
AccuSense™ Pressure Transducer, 0-100 PSIG
AccuSense™ Pressure Transducer, 0-100 PSIA
AccuSense™ Pressure Transducer, 0-150 PSIG
AccuSense™ Pressure Transducer, 0-250 PSIG.
More Details
Accuracy of Ph Meters
Buy Accuracy of Ph Meters Items Including:
Cole-Parmer High-Accuracy pH Electrode, 10-ft Cable
Cole-Parmer High-Accuracy pH Electrode, 100Ω Pt RTD, 10-ft Cable
Cole-Parmer High-Accuracy pH Electrode, 1 kΩ Pt RTD, 10-ft Cable
Cole-Parmer High-Accuracy pH Electrode, 100Ω Pt RTD, 25-ft Cable.
More Details
Accuracy of T Type Thermocouple
Buy Accuracy of T Type Thermocouple Items Including:
Cole-Parmer USB Data Acquisition Module, 8 channel thermocouple (USB-TC).
More Details
Accurate Balance Lab Scales Granger
Buy Accurate Balance Lab Scales Granger Items Including:
Ohaus Software, Triple Beam Virtual Lab, Single User.
More Details
Accurate Dial Vacuum Gauge
Buy Accurate Dial Vacuum Gauge Items Including:
Vacuum Gauge with Digital Dial and Reading (682,902.0)
Winters Tube Stub Gauge, Stainless Steel, 2 1/2" Dial, Vacuum
Vacuum Meter/Controller with Dogotal Dial and Reading, 810 to 0.1 Torr
Quality Process Gauge, 0 to 200 psi - 2 1/2" dial.
More Details
Accurate Digital Tire Pressure Gauge
Buy Accurate Digital Tire Pressure Gauge Items Including:
Digital Pressure Gauge, 0-100 PSI
Digital Pressure Gauge, 0-200 PSI
Digital Pressure Gauge, 0-300 PSI
Digital Pressure Gauge, 0-500 PSI
Digital Pressure Gauge, 0-1000 PSI
More Details
Accurate Organic Bottle Top Dispenser
Buy Accurate Organic Bottle Top Dispenser Items Including:
Cole-Parmer Bottletop Dispenser .25-2.5 mL
Cole-Parmer Bottletop Dispenser .25-2.5 mL with 100 mL Glass Bottle
Cole-Parmer Bottletop Dispenser 2.5-30 mL with 500 mL Glass Bottle
Cole-Parmer Bottletop Dispenser 5-60 mL with 1000 mL Glass Bottle.
More Details
Accurate Outdoor Temperature Thermometer
Buy Accurate Outdoor Temperature Thermometer Items Including:
12" Vertical Metal Wall Thermometer with Alcohol Spirit Tube
-50F to
12" Vertical Metal Wall Thermometer with Alcohol Spirit Tube
0F to 12
Thermometers with Time/Date and Wire Probe (4105CP)
More Details
Accurate Soil Ph Meter
Buy Accurate Soil Ph Meter Items Including:
Soil pH Electrode, BNC Connector (YO-05992-62)
Soil pH/conductivity meter kit (AG6/PH/PSTK)
Horiba Compact Nitrate Pocket Tester, for soil
Soil sampler extension rod, 26" long (PN004)
Replacement reagents for soil texture test kit (R-1067).
More Details
Accurate Thermometer Temperature Reading Armpit
Buy Accurate Thermometer Temperature Reading Armpit Items Including:
Easy-read General Purpose Thermometer, 20/230°F, 200mm Length, 50m
Easy-read General Purpose Thermometer, 20/230°F, 200mm Length, Tot
Easy-read General Purpose Thermometer, 20/300°F, 200mm Length, 50m.
More Details
Accurate Water Pressure Gauges
Buy Accurate Water Pressure Gauges Items Including:
Pressure Gauge ~ 1/4" NPT(M) process connection; 0 to 10" Water Co
Pressure Gauge ~ 1/4" NPT(M) process connection; 0 to 15" Water Co
Pressure Gauge ~ 1/4" NPT(M) process connection; 0 to 30" Water Co.
More Details
Accurate Wireless Rain Gauge
Buy Accurate Wireless Rain Gauge Items Including:
General Wireless Rain Gauge
Rainwise Wireless Dual Display Rain Gauge.
More Details
Accusense High-Accuracy Pressure Transducers
Buy Accusense High-Accuracy Pressure Transducers Items Including:
AccuSense™ Pressure Transducer, 30-0" HG
AccuSense™ Pressure Transducer, 0-100 PSIG
AccuSense™ Pressure Transducer, 0-100 PSIA
AccuSense™ Pressure Transducer, 0-150 PSIG
AccuSense™ Pressure Transducer, 0-250 PSIG.
More Details
Accutherm Non Mercury Astm Thermometers
Buy Accutherm Non Mercury Astm Thermometers Items Including:
Thermo Scientific ERTCO® Non-Mercury ASTM Glass Thermometer, ASTM (062CBBLSFC).
More Details
Ace G Funnels
Buy Ace G Funnels Items Including:
ACE GLASS Separatory Funnels, 125 mL (YO-34731-02)
ACE GLASS Separatory Funnels, 250 mL (YO-34731-04)
ACE GLASS Separatory Funnels, 1000 mL (YO-34731-08)
ACE GLASS Separatory Funnels, 2000 mL (YO-34731-10).
More Details
Ace G Separatory Funnels
Buy Ace G Separatory Funnels Items Including:
ACE GLASS Separatory Funnels, 125 mL (YO-34731-02)
ACE GLASS Separatory Funnels, 250 mL (YO-34731-04)
ACE GLASS Separatory Funnels, 500 mL (YO-34731-06)
ACE GLASS Separatory Funnels, 1000 mL (YO-34731-08).
More Details
Ace Glass
Buy Ace Glass Items Including:
ACE GLASS Separatory Funnels, 125 mL (YO-34731-02)
ACE GLASS Separatory Funnels, 250 mL (YO-34731-04)
ACE GLASS Separatory Funnels, 1000 mL (YO-34731-08)
Funnel, Ace, Powder Offset, 75MM (YO-34732-00)
Ace Glass Tube Adapter, 24/40 Joint (5028-30).
More Details
Ace Glass Centrifuge Tubes with Filter
Buy Ace Glass Centrifuge Tubes with Filter Items Including:
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 5 ml, 125 (99502-5)
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 10 ml, 12 (99502-10)
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 15 ml, 12 (99502-15).
More Details
Ace Glass Funnels
Buy Ace Glass Funnels Items Including:
ACE GLASS Separatory Funnels, 125 mL (YO-34731-02)
ACE GLASS Separatory Funnels, 250 mL (YO-34731-04)
ACE GLASS Separatory Funnels, 1000 mL (YO-34731-08)
ACE GLASS Separatory Funnels, 2000 mL (YO-34731-10).
More Details
Ace Glass Laboratory Supplies & Consumables
Buy Ace Glass Laboratory Supplies & Consumables Items Including:
ACE GLASS Separatory Funnels, 125 mL (YO-34731-02)
ACE GLASS Separatory Funnels, 250 mL (YO-34731-04)
ACE GLASS Separatory Funnels, 1000 mL (YO-34731-08)
Funnel, Ace, Powder Offset, 75MM (YO-34732-00)
Ace Glass Tube Adapter, 24/40 Joint (5028-30).
More Details
Ace Glass Separatory Funnels
Buy Ace Glass Separatory Funnels Items Including:
ACE GLASS Separatory Funnels, 125 mL (YO-34731-02)
ACE GLASS Separatory Funnels, 250 mL (YO-34731-04)
ACE GLASS Separatory Funnels, 500 mL (YO-34731-06)
ACE GLASS Separatory Funnels, 1000 mL (YO-34731-08).
More Details
Ace Glass Tube Adapter
Buy Ace Glass Tube Adapter Items Including:
Ace Glass Tube Adapter with Hose Barb, 24/40 Joint (5261-16).
More Details
Ace Mini Disposable Stir Bars
Buy Ace Mini Disposable Stir Bars Items Including:
1/2" X 1/8" Disposable Stir Bars (80037-1065)
1" X 5/16" Disposable Stir Bars (80037-1066)
1-1/2" X 5/16" Disposable Stir Bars (80037-1067).
More Details
Acetal Check Valves
Buy Acetal Check Valves Items Including:
Acetal Check Valve, 1/4" NPT(F) inlet/1/4" NPT(F) outlet (0354300)
Acetal Check Valve, 1/4" NPT(F) inlet/1/4" NPT(M) outlet (0354303)
CHECK VALVE, ACETAL BODY, push-to connect, 5/16'' tube OD (A5VC5-MG).
More Details
70618
results
<<<Previous
16
17
18
19
20
21
22
23
24
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Accumetric to Acetal Check Valves
>
31125329-s-toc_2 31125329stoc2