Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 21
Acetal Manifolds
to
Acetone Reagent
Image
Product Detail
More Detail
Acetal Manifolds
Buy Acetal Manifolds Items Including:
Acetal manifold, 2 port 1/4" NPT(F) inlet, 1/8" NPT(F) outlet (YO-31524-00)
Acetal manifold, 2 port 3/8" NPT(F) inlet, 1/4" NPT(F) outlet (YO-31524-02).
More Details
Acetal Quick Disconnect Coupling
Buy Acetal Quick Disconnect Coupling Items Including:
Miniature Quick-Disconnect Coupling; Acetal; Straight-through, 1/16" I (SMC01)
Miniature Quick-Disconnect Coupling; Acetal; Straight-through, 1/8" ID (SMC02)
Quick-Disconnect Panel-Mount Hose Barb Fitting Body; Straight-through, (PMC1601).
More Details
Acetal Quick Disconnect Coupling Plug
Buy Acetal Quick Disconnect Coupling Plug Items Including:
Quick Disconnect Air Plug, 1/4 in NPT(F) (MP-1/4F)
Quick Disconnect Air Plug, 3/8 in NPT(F) (MP-3/8F)
Miniature Quick-Disconnect Coupling; Acetal; Valved, 1/16" ID (SMCD01)
Miniature Quick-Disconnect Coupling; Acetal; Valved, 3 mm ID (SMCDM3).
More Details
Acetal Quick Disconnect Fittings
Buy Acetal Quick Disconnect Fittings Items Including:
Quick-Disconnect Panel-Mount Hose Barb Fitting Body; Straight-through, (PLC16004)
Quick-Disconnect Panel-Mount Hose Barb Fitting Body; Straight-through, (PLC16005)
Quick-Disconnect Panel-Mount Hose Barb Fitting Body; Straight-through, (PLC16006).
More Details
Acetal Quick-Disconnect
Buy Acetal Quick-Disconnect Items Including:
Quick-Disconnect Panel-Mount Hose Barb Fitting Body; Straight-through, (PLC16004)
Quick-Disconnect Panel-Mount Hose Barb Fitting Body; Straight-through, (PLC16005)
Quick-Disconnect Panel-Mount Hose Barb Fitting Body; Straight-through, (PLC16006).
More Details
Acetal Tube
Buy Acetal Tube Items Including:
150 mL tube top vacuum filters, 0.22 µm, 12/cs (430320)
Prepared media tube, Acetate differential—prepared slant tube,
Coupling Insert, Two-Line, Acetal, 1/16" ID Tube (PTC22010)
Coupling Insert, Two-Line, Acetal, 1/8" ID Tube (PTC22020).
More Details
Acetal Tube Fitting
Buy Acetal Tube Fitting Items Including:
Coupling Body, Valved, 10mm OD Tube, 10/pack (APCD130M10SH)
Coupling Body, Valved, 1/4" ID Tube, 10/pack (APCD17004SH)
Coupling Body, Valved, 3/8" ID Tube, 10/pack (APCD17006SH)
Coupling Body, Panel Mount, 8mm OD Tube, 10/pack (APC120M8).
More Details
Acetate Clear
Buy Acetate Clear Items Including:
Acetate Faceshield, Clear
Acetate Faceshield, Clear, Anti Fog.
More Details
Acetate Face Shield Chemical Resistance
Buy Acetate Face Shield Chemical Resistance Items Including:
North Silver Shield Gloves, Small, pack of 10 pair
North Silver Shield Gloves, medium, pack of 10 pair
North Silver Shield Gloves, large, pack of 10 pair
Acetate Faceshield, Clear, Anti Fog
U.S
More Details
Acetate Face Shield with Headpiece
Buy Acetate Face Shield with Headpiece Items Including:
Acetate Faceshield, Clear
Acetate Faceshield, Clear, Anti Fog
U.S
Safety Double Matrix™ Shield Window, Clear, Acetate
Laboratory faceshield, single crown w/acetate shield.
More Details
Acetate Face Shields
Buy Acetate Face Shields Items Including:
Acetate Faceshield, Clear
Acetate Faceshield, Clear, Anti Fog
U.S
Safety Double Matrix™ Shield Window, Clear, Acetate
Laboratory faceshield, single crown w/acetate shield.
More Details
Acetate Faceshields
Buy Acetate Faceshields Items Including:
Acetate Faceshield, Clear
Acetate Faceshield, Clear, Anti Fog
Laboratory faceshield, single crown w/acetate shield.
More Details
Acetate Filters
Buy Acetate Filters Items Including:
Cole-Parmer Cellulose Acetate Sterile Filters; pore size; 0.45µ; Ac
Thermo Scientific Nalgene Surfactant-Free Cellulose Acetate (SFCA) Fil (YO-06731-20)
Thermo Scientific Nalgene Surfactant-Free Cellulose Acetate (SFCA) Fil (YO-06731-22).
More Details
Acetic Acid
Buy Acetic Acid Items Including:
Sensidyne Gas Detection Tubes, Acetic acid
CMS Chip, Acetic Acid, 2 to 50 ppm
Drager Tubes, Acetic Acid, 5 To 80 ppm
Drager Diffusion Tube Acetic Acid 10/a-D
ACETIC ACID GLACIAL ACS 2 1/2L (YO-88051-42)
More Details
Acetic Acid 06 3 Ml
Buy Acetic Acid 06 3 Ml Items Including:
C18, water wettable SPE cartridges, 3 mL (YO-10937-06)
Si-propylsulfonic acid SPE cartridges, 3 mL (YO-10937-36)
Push-to-connect stem-to-hose adapters, 5/16" x 3/16", 10/pack (YO-34007-06)
2 3 -DIMETHYLBENZOIC ACID 5G (YO-88282-06).
More Details
Acetic Acid 10 Solution
Buy Acetic Acid 10 Solution Items Including:
Drager Diffusion Tube Acetic Acid 10/a-D
Drager Tubes, Acetic Acid, 5 To 80 ppm.
More Details
Acetic Acid 2 Ear Solution
Buy Acetic Acid 2 Ear Solution Items Including:
ACETIC ACID GLACIAL ACS 2 1/2L (YO-88051-42).
More Details
Acetic Acid Atlanta Ga
Buy Acetic Acid Atlanta Ga Items Including:
Sensidyne Gas Detection Tubes, Acetic acid
Drager Tubes, Acetic Acid, 5 To 80 ppm
CMS Chip, Acetic Acid, 2 to 50 ppm.
More Details
Acetic Acid Chemical Warehouse
Buy Acetic Acid Chemical Warehouse Items Including:
ACETIC ACID GLACIAL ACS 2 1/2L (YO-88051-42)
GLAC ACETIC ACID USPFCCEPBP2.5 (YO-88053-31)
ACETIC ACID GLACIAL ACS GRADE (YO-88058-60)
ACETIC ACID POTASSIUM SAL 1KG (YO-88235-45)
ACETIC ACID POTASSIUM SA 100G (YO-88235-46).
More Details
Acetic Acid Glacial
Buy Acetic Acid Glacial Items Including:
ACETIC ACID GLACIAL ACS 2 1/2L (YO-88051-42)
ACETIC ACID GLACIAL ACS PLUS (YO-88058-57).
More Details
Acetic Acid Glacial Acs
Buy Acetic Acid Glacial Acs Items Including:
ACETIC ACID GLACIAL ACS 2 1/2L (YO-88051-42)
ACETIC ACID GLACIAL ACS PLUS (YO-88058-57)
ACETIC ACID GLACIAL ACS GRADE (YO-88058-60).
More Details
Acetic Acid Glacial Acs Grade
Buy Acetic Acid Glacial Acs Grade Items Including:
ACETIC ACID GLACIAL ACS GRADE (YO-88058-60)
ACETIC ACID GLACIAL ACS 2 1/2L (YO-88051-42)
ACETIC ACID GLACIAL ACS PLUS (YO-88058-57).
More Details
Acetic Acid Products
Buy Acetic Acid Products Items Including:
Sensidyne Gas Detection Tubes, Acetic acid
CMS Chip, Acetic Acid, 2 to 50 ppm
Drager Tubes, Acetic Acid, 5 To 80 ppm
Drager Diffusion Tube Acetic Acid 10/a-D
ACETIC ACID GLACIAL ACS 2 1/2L (YO-88051-42)
More Details
Acetic Acid, Sodium Salt
Buy Acetic Acid, Sodium Salt Items Including:
ACETIC ACID SODIUM SALT 500GR (YO-88362-67).
More Details
Acetic Anhydride
Buy Acetic Anhydride Items Including:
ACETIC ANHYDRIDE REAGENT 1 LT (YO-88360-45).
More Details
Acetone 30 Ml Bottle
Buy Acetone 30 Ml Bottle Items Including:
30 mL Bottle and glass dropper (15040G-30)
Thermo Scientific Nalgene polypropylene wide-mouth bottle, 30 mL
30 mL Kimax Glass Dropping Bottle
30 mL Amber Glass Dropping Bottle, Plastic dropper
Azlon solvent venting wash bottle for acetone, 250 mL.
More Details
Acetone 55 Gallon Drum
Buy Acetone 55 Gallon Drum Items Including:
4in Threaded Standoff Kit For 55 Gallon Drums
12in Threaded Standoff Kit For 55 Gallon Drums
55 Gallon Drum Insulation Cover (for Top Of 55 Gallon Drums) (FGDC55)
DrumQuik PUR Drum Dip Tube for 55-gallon drum, 985 mm length, HDPE, pa (YO-31608-73).
More Details
Acetone Acs
Buy Acetone Acs Items Including:
ACETONE ACS 4 LITER SAFE-TINTM (YO-88050-60)
ACETONE ACS GRADE (YO-88058-63)
ACETONE ACS GRADE (YO-88058-64)
CONTEC Prewetted Cleanroom Wipes, 100% acetone
ACETONE CERTIFIED ACS 20L (YO-88050-37)
More Details
Acetone Hplc Grade Acs Reagent Grade
Buy Acetone Hplc Grade Acs Reagent Grade Items Including:
ACETONE ACS GRADE (YO-88058-63).
More Details
Acetone Reagent
Buy Acetone Reagent Items Including:
ACETONE ACS GRADE (YO-88058-63)
ACETONE ACS GRADE (YO-88058-64)
ACETONE NF/FCC 1LITER (YO-88050-14)
ACETONE NF/FCC 200 LITER (YO-88050-20)
ACETONE NF/FCC 4 LITER (YO-88050-21)
More Details
70618
results
<<<Previous
17
18
19
20
21
22
23
24
25
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
40
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Acetal Manifolds to Acetone Reagent
>
31125329-s-toc_2 31125329stoc2