Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 22

Acetone Reagent Acs Usp Nf Grade   to   Acid Storage Cabinets

Image
  Product Detail
More Detail

Acetone Reagent Acs Usp Nf Grade

Buy Acetone Reagent Acs Usp Nf Grade Items Including:
  • ACETONE ACS GRADE (YO-88058-63).

Acetone Solvent

Buy Acetone Solvent Items Including:
  • Azlon solvent venting wash bottle for acetone, 250 mL.

Acetone Sterile Liquid Filter

Buy Acetone Sterile Liquid Filter Items Including:
  • Cole-Parmer Filter Pipette Tips, sterile, 2" L, 10
  • Finntip Flex Filter Pipette Tips, 100-1000 uL, non-sterile, 1000/bag (94052440)
  • Filter non-sterile 5/pk (4580560)
  • Replacement filters, 0.45µm non-sterile for Omega Pipettors, 10/pk (153 016).

Acetone Wash

Buy Acetone Wash Items Including:
  • Azlon solvent venting wash bottle for acetone, 250 mL
  • Azlon solvent venting wash bottle for acetone, 500 mL
  • Scienceware safety labeled 500-mL wash bottle for acetone
  • Multi-Language Wash Bottle, LDPE Acetone, 250 mL 5/pk.

Acetone Wash Bottle

Buy Acetone Wash Bottle Items Including:
  • Azlon solvent venting wash bottle for acetone, 250 mL
  • Azlon solvent venting wash bottle for acetone, 500 mL
  • Scienceware safety labeled 500-mL wash bottle for acetone
  • Multi-Language Wash Bottle, LDPE Acetone, 250 mL 5/pk.

Acetonitrile Acs

Buy Acetonitrile Acs Items Including:
  • ACETONITRILE CERTIFIED ACS 20L (YO-88050-72).

Acetonitrile Hplc

Buy Acetonitrile Hplc Items Including:
  • ACETONITRILE HPLC GRADE (YO-88058-72).

Achat Ball Mill Test Equipment

Buy Achat Ball Mill Test Equipment Items Including:
  • IKA ULTRA TURRAX Ball Mill Tube 50 mL, Glass (10/pk)
  • IKA ULTRA TURRAX Ball Mill Tube BMT-20 G Tube, (25/pk)
  • IKA ULTRA TURRAX Ball Mill Tube BMT-20 S Tube, (25/pk)
  • Stainless Steel Balls, 5 mm (YO-04182-59)
  • Stainless Steel Balls, 7 mm (YO-04182-60).

Acid 2.0

Buy Acid 2.0 Items Including:
  • SciMatCo HDPE Benchtop Acid Storage Cabinet, 30 x 2.5 L bottle capacit
  • SciMatCo Nitric Acid Compartment for 81780-20, -30
  • Micro A07 Citric Acid Cleaner 20L bottle (YO-18104-30)
  • Specialty 3M Particulate Disposable Respirators, R95, Standard, acid g.

Acid Base Indicator Methylene Blue

Buy Acid Base Indicator Methylene Blue Items Including:
  • Acid Spot'r™ Acid Detecting Spray, 16oz Trigger Spray (AA-002-01)
  • On Guard Acid Detecting Paint, Gallon (AA-001-01)
  • On Guard Rty Acid Neutralizer (AA-003-01).

Acid Base Ph Meter

Buy Acid Base Ph Meter Items Including:
  • Acid adjuster (977011)
  • Acid Spot'r™ Acid Detecting Spray, 16oz Trigger Spray (AA-002-01)
  • On Guard Acid Detecting Paint, Gallon (AA-001-01)
  • On Guard Rty Acid Neutralizer (AA-003-01)
  • Weighted base with 5 solution cups

Acid Base Titration Equipment

Buy Acid Base Titration Equipment Items Including:
  • Acid-Base titrator cartridge with range of 1 to 10 meq/L Acid
  • Titran (14379-01)
  • Aquamax Total Acid Number (TAN) Titrator
  • Metter Toledo DL15 Titrator
  • Alkalinity titrator cartridge with range of 10 to 160 mg/L CaCO3 and t (14388-01).

Acid Bottles

Buy Acid Bottles Items Including:
  • SciMatCo Full-Size Acid Storage Cabinet, twenty four 2.5 L bottle capa
  • SciMatCo HDPE Benchtop Acid Storage Cabinet, six 2.5 L bottle capacity
  • SciMatCo Polypropylene Acid Storage Cabinet, 6 X 2.5 L bottle capacity.

Acid Cabinets

Buy Acid Cabinets Items Including:
  • Justrite Solid Polyethylene Acid Cabinet, Undercounter version
  • Eagle Acid Storage Cabinet, Manual-Latching Door, 30 Gallon
  • Eagle Acid Storage Cabinet, Manual-Latching Door, 45 Gallon
  • Eagle Benchtop Acid Storage Cabinet, Self-Closing Door, 4 Gallon.

Acid Chemical Pumps

Buy Acid Chemical Pumps Items Including:
  • Pump Skid, Acid/base, With Calibration Column, 12 Gpd, 115 VAC
  • Pump Skid, Acid/base, With Calibration Column, 12 Gpd, 230 VAC
  • Pump Skid, Acid/base, With Calibration Column, 22 Gpd, 115 VAC.

Acid Chemicals

Buy Acid Chemicals Items Including:
  • Pump Skid, Acid/base, With Calibration Column, 12 Gpd, 115 VAC
  • Pump Skid, Acid/base, With Calibration Column, 12 Gpd, 230 VAC
  • Pump Skid, Acid/base, With Calibration Column, 22 Gpd, 115 VAC
  • KIT SPILL CLEANUP ACID/C & SOL (YO-88050-03).

Acid Cleaner

Buy Acid Cleaner Items Including:
  • Micro A07 Citric Acid Cleaner 1L bottle (YO-18104-00)
  • Micro A07 Citric Acid Cleaner case of 12 1L bottles (YO-18104-01)
  • Micro A07 Citric Acid Cleaner 20L bottle (YO-18104-30).

Acid Corrosive Cabinet

Buy Acid Corrosive Cabinet Items Including:
  • Eagle PE Acid And Corrosive Cabinets, 22 Gal, Blue
  • Eagle PE Acid And Corrosive Cabinets, 44 Gal, Blue
  • Eagle PE Acid And Corrosive Cabinets, 4 Gal, White
  • Eagle PE Acid And Corrosive Cabinets, 22 Gal, White.

Acid Digestion Laboratory Fume Hoods

Buy Acid Digestion Laboratory Fume Hoods Items Including:
  • Complete 4foot Fume Hood System With Acid Storage Cabinet Epoxy Worksu (YO-41525-00)
  • Complete 5foot Fume Hood System With Acid And Base Storage Cabinets Ep (YO-41525-10)
  • Labconco Fiberglass 30 fume hood, 1/10 HP blower, 115V (YO-09066-30).

Acid Eater

Buy Acid Eater Items Including:
  • Ultimate Acid Eater; 2.5 Gal.; 2 Packs/case
  • Ultimate Acid Eater Spill Kit; 5 Gal.
  • Hf Acid Eater; 2.5 Gal.; 2 Packs/case.

Acid Eater Spill Kit

Buy Acid Eater Spill Kit Items Including:
  • Ultimate Acid Eater Spill Kit; 5 Gal.
  • Hf Acid Eater Spill Kit; 5 Gal.; 1 Pack/case
  • Ultimate Acid Eater; 2.5 Gal.; 2 Packs/case
  • Hf Acid Eater; 2.5 Gal.; 2 Packs/case
  • SPILL-X-A Acid Agent Refill, 6 containers

Acid Extraction

Buy Acid Extraction Items Including:
  • Si-propylsulfonic acid SPE cartridges, 3 mL (YO-10937-36).

Acid Fume Hoods

Buy Acid Fume Hoods Items Including:
  • Base storage cabinet for acids for 30" fume hood (YO-09115-04)
  • Primary Mineral acid Filters for 30" Self-Monitoring Ductless Fume Hoo (YO-33713-60)
  • Primary Mineral acid Filters for 54" Self-Monitoring Ductless Fume Hoo (YO-33713-62).

Acid Ph Test Strips

Buy Acid Ph Test Strips Items Including:
  • Test Strips, Peracetic acid, 0-160ppm, 50 strips/vial (3000)
  • Acid Spot'r™ Acid Detecting Spray, 16oz Trigger Spray (AA-002-01)
  • On Guard Rty Acid Neutralizer (AA-003-01)
  • Liquid Sanitizer Strength Verification Kits, Peracetic Acid Test (7191-01).

Acid Pumps

Buy Acid Pumps Items Including:
  • Pump, Motorized Drum Pump Package, Designed for Acids and Alkalies, 35 (9400-110V) (YO-70614-02)
  • Pump, Motorized Drum Pump Package, Designed for Acids and Alkalies, 35 (9500-110V)
  • Pump Skid, Acid/base, With Calibration Column, 6 Gpd, 115 VAC.

Acid Refractometers

Buy Acid Refractometers Items Including:
  • Reichert Coolant/Battery Acid Refractometer; Ethylene Glycol: -60 to 3
  • Reichert Coolant/Battery Acid Refractometer; Ethylene Glycol: -48 to 0
  • Reichert Industrial Fluid Refractometer, 0 to 10 scale (1.3330 to 1.34.

Acid Resistant Peristaltic Pump Tubing

Buy Acid Resistant Peristaltic Pump Tubing Items Including:
  • Tube Assembly Kit for the peristaltic pump 74130-60 and 74130-61 pumps
  • Masterflex two-stop pump tubing, Tygon, L/S 15
  • Pack of 4.
  • Masterflex two-stop pump tubing, Tygon, L/S 24
  • Pack of 4.

Acid Spill Kits

Buy Acid Spill Kits Items Including:
  • SPILL-X-A Acid Spill Treatment Kit
  • SPILL-X-A Acid Agent Refill, 6 containers
  • Ultimate Acid Eater Spill Kit; 5 Gal.
  • Hf Acid Eater Spill Kit; 5 Gal.; 1 Pack/case
  • KIT ACID SPILL CLEAN-UP (YO-88050-00).

Acid Storage

Buy Acid Storage Items Including:
  • SciMatCo Full-Size Acid Storage Cabinet, twenty four 2.5 L bottle capa
  • SciMatCo Full-Size Acid Storage Cabinet with Full Lining, thirty 2.5 L
  • SciMatCo HDPE Benchtop Acid Storage Cabinet, six 2.5 L bottle capacity.

Acid Storage Cabinets

Buy Acid Storage Cabinets Items Including:
  • SciMatCo Full-Size Acid Storage Cabinet, twenty four 2.5 L bottle capa
  • SciMatCo Full-Size Acid Storage Cabinet with Full Lining, thirty 2.5 L
  • SciMatCo HDPE Benchtop Acid Storage Cabinet, six 2.5 L bottle capacity.



Acetone Reagent Acs Usp Nf Grade to Acid Storage Cabinets

>