Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 25

Acs Grade   to   Adam Abk Industrial Scales

Image
  Product Detail
More Detail

Acs Grade

Buy Acs Grade Items Including:
  • ACETIC ACID GLACIAL ACS GRADE (YO-88058-60)
  • ACETONE ACS GRADE (YO-88058-63).

Acs Grade Acetone

Buy Acs Grade Acetone Items Including:
  • ACETONE ACS GRADE (YO-88058-63).

Acs Reagent

Buy Acs Reagent Items Including:
  • CHROMIUM TRIOX CERT ACS 21/2KG (YO-88050-06)
  • CITRIC ACID GRAN CERT ACS 10KG (YO-88050-07)
  • CITRIC AC MONOHYD USP/FCC 10KG (YO-88050-11)
  • CITRIC AC MONOHYD USP/FCC 3KG (YO-88050-12)
  • PERCHLORIC AC 60% ACS 4X7LB/CS (YO-88051-01).

Acs Reagent Grade Sodium Hydroxide

Buy Acs Reagent Grade Sodium Hydroxide Items Including:
  • Sodium Hydroxide Solution (0.2N (N/5)/Certified) (1 liter) (YO-88024-44)
  • SODIUM HYDROXIDE REAGENT 25G (YO-88362-85)
  • SODIUM HYDROXIDE NF/EP/BP/FCC (YO-88081-49)
  • SODIUM HYDROXIDE REAGEN 2.5KG (YO-88362-83)
  • SODIUM HYDROXIDE 50 WT% 25ML (YO-88255-51).

Acsi Digital Pressure Gauge

Buy Acsi Digital Pressure Gauge Items Including:
  • Ashcroft Digital Pressure Gauge, 0 to 1000 psi, 1/4" NPT(M) Process co (D1005PS2)
  • Additel Digital Pressure Gauge, 0" to 30" Hg, 1/4" NPT(M) Connection
  • Additel Digital Pressure Gauge, 0 to 100 psi, 1/4" NPT(M) Connection.

Action Back Coveralls

Buy Action Back Coveralls Items Including:
  • Action back coveralls; Size, medium; chest size, 38" to 40"; color, na
  • Action back coveralls; size, large; chest size, 42" to 44"; color, nav
  • Action back coveralls; size, X-large; chest size, 46" to 48"; color, n.

Action Instruments

Buy Action Instruments Items Including:
  • I/q Rail AC Pwr Dist Asy 2mod
  • I/q Rail DC Pwr Dist Asy 4mod
  • I/q Rail AC Pwr Dist Asy 4mod
  • Davis Item Actq50113000
  • I/q Rail DC Pwr Dist Asy 8mod.

Action Instruments Air Cooling

Buy Action Instruments Air Cooling Items Including:
  • I/q Rail AC Pwr Dist Asy 2mod
  • I/q Rail DC Pwr Dist Asy 4mod
  • I/q Rail AC Pwr Dist Asy 4mod
  • Davis Item Actq50113000.

Action Instruments Environmental Testing

Buy Action Instruments Environmental Testing Items Including:
  • I/q Rail AC Pwr Dist Asy 2mod
  • I/q Rail DC Pwr Dist Asy 4mod
  • I/q Rail AC Pwr Dist Asy 4mod
  • Davis Item Actq50113000.

Action Instruments Shop by Industry

Buy Action Instruments Shop by Industry Items Including:
  • I/q Rail AC Pwr Dist Asy 2mod
  • I/q Rail DC Pwr Dist Asy 4mod
  • I/q Rail AC Pwr Dist Asy 4mod
  • Davis Item Actq50113000.

Activate &

Buy Activate & Items Including:
  • LabSwift Portable Water Activity Instrument
  • LabTouch Water Activity Instrument, with Semi-Temp Stabilized
  • LabMaster Standard Water Activity Instrument, with Full Temperature Co
  • LabMaster AdvancedWater Activity Instrument, with Full Temperature Con.

Activated Carbon Air Capsule Filter

Buy Activated Carbon Air Capsule Filter Items Including:
  • Double open end granular activated carbon cartridge; 20 µm, 10"L (155155-43)
  • Double open end granular activated carbon cartridge; 20 µm, 20"L (155138-43)
  • Parker Domnick Hunter Compressed Air Activated Carbon Filter Element, (YO-29587-21).

Activated Carbon Capsule Filters

Buy Activated Carbon Capsule Filters Items Including:
  • Double open end granular activated carbon cartridge; 20 µm, 10"L (155155-43)
  • Activated Carbon After-Filter
  • Double open end activated carbon-impregnated paper cartridge; 5 µm, (155002-03)
  • Activated Carbon Refill for 33703-04, -05 (YO-33703-07).

Activated Carbon Cartridges

Buy Activated Carbon Cartridges Items Including:
  • Double open end granular activated carbon cartridge; 20 µm, 10"L (155155-43)
  • Double open end granular activated carbon cartridge; 20 µm, 20"L (155138-43)
  • Double open end activated carbon-impregnated paper cartridge; 5 µm, (155002-03).

Activated Carbon Filter Paper

Buy Activated Carbon Filter Paper Items Including:
  • Double open end activated carbon-impregnated paper cartridge; 5 µm, (155002-03).

Activated Carbon Filters

Buy Activated Carbon Filters Items Including:
  • Double open end granular activated carbon cartridge; 20 µm, 10"L (155155-43)
  • Double open end granular activated carbon cartridge; 20 µm, 20"L (155138-43)
  • Parker Domnick Hunter Compressed Air Activated Carbon Filter Element, (YO-29587-21).

Activated Carbon Media

Buy Activated Carbon Media Items Including:
  • Filter cartridge, "Big Blue", 10", carbon block, 0.5 micron (155170-43(CBC-BB))
  • Filter cartridge, "Big Blue", 20", carbon block, 0.5 micron (155312-43(CBC-20BB)).

Active Probe

Buy Active Probe Items Including:
  • TekVPI 500 MHz differential active probe (TDP0500)
  • Handheld Meter Kit, 14-mm sample cup size, one probe input (one probe (HYGROPALM-AW1 W/14ML CUP)
  • Handheld Meter Kit, 40-mm sample cup size, one probe input (one probe (HYGROPALM-AW1 W/40ML CUP).

Active Vacuum Switch/Transmitters

Buy Active Vacuum Switch/Transmitters Items Including:
  • Pirani Type Switch/transmitter, 10-5 To 10-1 Tor
  • Thermocouple Switch/transmitter, 1 To 1000 Mtorr
  • Switch/transmitter, 3 To 5000 Mtorr
  • Diaphragm Type Switch/transmitter, 1 To 1500 Torr
  • Diaphragm Type Switch/transmitter, 30" Hg To Atmosphere.

Actron Pocket Infrared Thermometer

Buy Actron Pocket Infrared Thermometer Items Including:
  • Extech IR201 : Pocket IR Thermometer, 0/518F with Laser, Fixed Emissiv.

Actuated Ball Valve Pvc

Buy Actuated Ball Valve Pvc Items Including:
  • Hayward electrically Actuated Ball Valve, 3-way, 1" PVC (HCTN1100STV/EAU-28)
  • Hayward electrically Actuated Ball Valve, 3-way, 2" PVC (HCTN1200STV/EAU-28)
  • Hayward electrically Actuated Ball Valve, 3-way, 1/2" CPVC (HCTN2050STV/EAU-28).

Actuated Elliptical Valve Systems

Buy Actuated Elliptical Valve Systems Items Including:
  • Electrically Actuated 2-Position, 3-Way Elliptical Valve System; PP bo (223-PP-1-RE1)
  • Pneumatically Actuated 2-Way Elliptical Valve System; PP body, 3/8" NP (32-PP-RA4)
  • Two-Way Elliptical Valve, PP, 115 VAC, 1/8".

Acu Rite Refrigerator and Freezer Thermometer

Buy Acu Rite Refrigerator and Freezer Thermometer Items Including:
  • Digital-Bottle Refrigerator/Freezer Thermometer with solution filled b
  • Digital-Bottle High Accuracy Refrigerator/Freezer Thermometer with sol
  • Jumbo Refrigerator/freezer thermometer with a 5ml bottle that fits in (4627CP).

Acu Rite Wireless Rain Gauge

Buy Acu Rite Wireless Rain Gauge Items Including:
  • General Wireless Rain Gauge
  • Rainwise Wireless Dual Display Rain Gauge.

Acu Rite Wireless Weather Instruments

Buy Acu Rite Wireless Weather Instruments Items Including:
  • Davis Instruments Vantage Pro2™ Plus Wireless Weather Station with
  • General Instruments Professional Wireless Weather Station
  • Davis Instruments Vantage Vue Wireless LCD Console/Receiver
  • Davis Instruments Vantage Vue Wireless Integrated Sensor Suite.

Acumet Ph Meter

Buy Acumet Ph Meter Items Including:
  • acumet® XL 60 Multiparameter Meter with self-stirring BOD probe.

Acurite Outdoor Thermometer Humidity

Buy Acurite Outdoor Thermometer Humidity Items Including:
  • Indoor/Outdoor Thermometer with settable Hi/Low audible alarm
  • Indoor/Outdoor Thermometer (4126CP)
  • Indoor/Outdoor Thermometer with Alarm (7141CP)
  • Thermometers with Time/Date and Wire Probe (4105CP)
  • Thermometers with Time/Date and 5 ml Bottle Probe.

Acurite Wireless Weather Station

Buy Acurite Wireless Weather Station Items Including:
  • Davis Instruments Vantage Pro2™ Wireless Weather Station
  • Davis Instruments Vantage Pro2™ Plus Wireless Weather Station with
  • Davis Instruments Vantage Pro2™ Wireless Temperature Station
  • Davis Instruments Vantage Vue Wireless Integrated Sensor Suite.

Ada Chemical Fume Hood

Buy Ada Chemical Fume Hood Items Including:
  • 1 Drawer Wood Lab Table With Chemical Resistant Laminate Top 36in W X (YO-41527-50)
  • 2 Drawer Wood Lab Table With Chemical Resistant Laminate Top 48in W X (YO-41527-52).

Adam Abk Industrial Scales

Buy Adam Abk Industrial Scales Items Including:
  • Adam ABK Industrial Scale, 130lb/60kg Capacity and 0.005lb/2g Readabil (ABK 130A) (YO-11120-01)
  • Adam ABK Industrial Scale, 260lb/120kg Capacity and 0.01lb/5g Readabil (ABK 260A) (YO-11120-03).



Acs Grade to Adam Abk Industrial Scales

>