Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 26
Adam Afk Industrial Scales
to
Adam Equipment Balance Anti-Theft Devices
Image
Product Detail
More Detail
Adam Afk Industrial Scales
Buy Adam Afk Industrial Scales Items Including:
Adam AFK Industrial Scale, 165lb/75kg Capacity and 0.01lb/5g Readabili (AFK 165A) (YO-11120-04)
Adam AFK Industrial Scale, 165lb/75kg Capacity and 0.01lb/5g Readabili (AFK 165A) (YO-11120-06).
More Details
Adam Analytical Balances
Buy Adam Analytical Balances Items Including:
Adam PW Analytical Balance, 120 g x 0.0001 g, 220 VAC (PW124_220V)
Adam PGD 84e Analytical Balance, 80g with 0.1mg readability, graphic L (YO-11120-98)
Adam PGD 84e Analytical Balance, 80g with 0.1mg readability, graphic L (YO-11120-99).
More Details
Adam Balance Accessories
Buy Adam Balance Accessories Items Including:
Below Balance Hanger for Density Determination (7238)
Balance In-Use Wet Cover for Adam Equipment balances (7997)
Balance In-use wet cover, Large for Adam Equipment balances (8031)
Balance In-use wet cover, small for Adam Equipment balances (5111).
More Details
Adam Balance and Scale Accessories
Buy Adam Balance and Scale Accessories Items Including:
Density Determination Kit for Adam Balances
Precision Toploading Balance Dust Cover for Adam Balances
Kensington™-type security lock and cable (8030)
HAND PUMP (HP)
In use wet cover for CBD, CBC, CBW, CBK Qty
More Details
Adam Balances
Buy Adam Balances Items Including:
Precision Toploading Balance Dust Cover for Adam Balances
Printer paper for Adam Equipment printers, pack of 5 rolls (PRINTER PAPER)
Density Determination Kit for Adam Equipment balances (7543)
Below Balance Hanger for Density Determination (7238).
More Details
Adam Balances & Scales
Buy Adam Balances & Scales Items Including:
Precision Toploading Balance Dust Cover for Adam Balances
Printer paper for Adam Equipment printers, pack of 5 rolls (PRINTER PAPER)
Density Determination Kit for Adam Equipment balances (7543)
Below Balance Hanger for Density Determination (7238).
More Details
Adam Cbc Counting Scales
Buy Adam Cbc Counting Scales Items Including:
Adam CBC Counting Scale, 4 Kg X 0.1g
Adam CBC Counting Scale, 4 Kg X 0.1g, 220 VAC
Adam CBC Counting Scale, 8 Kg X 0.2g, 220 VAC
Adam CBC Counting Scale, 32 Kg X 1g
Adam CBC Counting Scale, 32 Kg X 1g, 220 VAC
More Details
Adam Cbk Compact Industrial Bench Scales
Buy Adam Cbk Compact Industrial Bench Scales Items Including:
Adam CBK Compact Industrial Bench Scale, 4 Kg X 0.1g, 220 VAC
Adam CBK Compact Industrial Bench Scale, 8 Kg X 0.1g, 220 VAC
Adam CBK Compact Industrial Bench Scale, 16 Kg X 0.5g, 220 VAC
Adam CBK Compact Industrial Bench Scale, 32 Kg X 1g, 220 VAC.
More Details
Adam Core Portable Balances
Buy Adam Core Portable Balances Items Including:
Adam Core™ Portable Balance, 200g X 0.01g
Adam Core™ Portable Balance, 250g X 0.1g
Adam Core™ Portable Balance, 600g X 0.1g
Adam Core™ Portable Balance, 1500g X 0.1g
Adam Core™ Portable Balance, 2000g X 1g
More Details
Adam Cpw Plus-L Industrial Scales
Buy Adam Cpw Plus-L Industrial Scales Items Including:
Adam CPW Plus-P Industrial Scale, 330lb/150kg Capacity and 0.1lb/50g R (CPWPLUS 150P) (YO-11120-47)
Adam CPW Plus-P Industrial Scale, 330lb/150kg Capacity and 0.1lb/50g R (CPWPLUS 150P) (YO-11120-48).
More Details
Adam Data Acquisition
Buy Adam Data Acquisition Items Including:
12 Channel Sourcing Iso Digital Output Data Aquisition Module (Modbus)
8 Channel Relay Output Module (Modbus) (ADAM-4068-BE)
Isolated RS-232 to RS-422/485 Converter (ADAM-4520-D2E)
2-port Ethernet to RS-232/485/422 ComPort Server (ADAM-4570-BE).
More Details
Adam Digital Health Scales (Mdw-250l/300l)
Buy Adam Digital Health Scales (Mdw-250l/300l) Items Including:
Adam Digital Health Scale, MDW-250L, 550lb/250kg Capacity 115V
Adam Digital Health Scale, MDW-250L, 550lb/250kg Capacity 230V
Adam Digital Health Scale, MDW-300L, 600lb/300kg Capacity 115V.
More Details
Adam Eclipse Toploading Balances
Buy Adam Eclipse Toploading Balances Items Including:
Adam PGD 253i Precision Toploader Balance, 250g w/1mg readability, gra (YO-11121-36)
Adam PGD 253i Precision Toploader Balance, 250g w/1mg readability, gra (YO-11121-37).
More Details
Adam Equipment
Buy Adam Equipment Items Including:
Carrying Case for Adam Equipment CPW Plus Industrial Scale
Balance In-use wet cover, Large for Adam Equipment balances (8031)
Density Determination Kit for Adam Balances
Balance In-use wet cover for Adam CBK, CBD, CBC, and CBW Series Balanc (5006).
More Details
Adam Equipment Abk Industrial Scales
Buy Adam Equipment Abk Industrial Scales Items Including:
Adam ABK Industrial Scale, 130lb/60kg Capacity and 0.005lb/2g Readabil (ABK 130A) (YO-11120-01)
Adam ABK Industrial Scale, 260lb/120kg Capacity and 0.01lb/5g Readabil (ABK 260A) (YO-11120-03).
More Details
Adam Equipment Adam Abk Industrial Scales
Buy Adam Equipment Adam Abk Industrial Scales Items Including:
Adam ABK Industrial Scale, 130lb/60kg Capacity and 0.005lb/2g Readabil (ABK 130A) (YO-11120-01)
Adam ABK Industrial Scale, 260lb/120kg Capacity and 0.01lb/5g Readabil (ABK 260A) (YO-11120-03).
More Details
Adam Equipment Adam Afk Industrial Scales
Buy Adam Equipment Adam Afk Industrial Scales Items Including:
Adam AFK Industrial Scale, 165lb/75kg Capacity and 0.01lb/5g Readabili (AFK 165A) (YO-11120-04)
Adam AFK Industrial Scale, 165lb/75kg Capacity and 0.01lb/5g Readabili (AFK 165A) (YO-11120-06).
More Details
Adam Equipment Adam Balance Accessories
Buy Adam Equipment Adam Balance Accessories Items Including:
Density Determination Kit for Adam Equipment balances (7543)
Density Determination Kit for Adam Balances
Balance In-use wet cover for Adam CBK, CBD, CBC, and CBW Series Balanc (5006)
Analytical Balance Dust Cover for Adam Equipment balances (8003).
More Details
Adam Equipment Adam Cbc Counting Scales
Buy Adam Equipment Adam Cbc Counting Scales Items Including:
Adam CBC Counting Scale, 4 Kg X 0.1g
Adam CBC Counting Scale, 4 Kg X 0.1g, 220 VAC
Adam CBC Counting Scale, 8 Kg X 0.2g, 220 VAC
Adam CBC Counting Scale, 32 Kg X 1g
Adam CBC Counting Scale, 32 Kg X 1g, 220 VAC
More Details
Adam Equipment Adam Core Portable Balances
Buy Adam Equipment Adam Core Portable Balances Items Including:
Adam Core™ Portable Balance, 200g X 0.01g
Adam Core™ Portable Balance, 250g X 0.1g
Adam Core™ Portable Balance, 600g X 0.1g
Adam Core™ Portable Balance, 1500g X 0.1g
Adam Core™ Portable Balance, 2000g X 1g
More Details
Adam Equipment Adam Eclipse Toploading Balances
Buy Adam Equipment Adam Eclipse Toploading Balances Items Including:
Adam PGD 253i Precision Toploader Balance, 250g w/1mg readability, gra (YO-11121-36)
Adam PGD 253i Precision Toploader Balance, 250g w/1mg readability, gra (YO-11121-37).
More Details
Adam Equipment Adam Equip Balance Accessories
Buy Adam Equipment Adam Equip Balance Accessories Items Including:
Density Determination Kit for Adam Balances
Kensington™-type security lock and cable (8030)
In use wet cover for CBD, CBC, CBW, CBK Qty
10
In use wet cover for CBD, CBC, CBW, CBK Qty
More Details
Adam Equipment Adam Gbk Industrial Scales
Buy Adam Equipment Adam Gbk Industrial Scales Items Including:
Adam GBK Industrial Scale, 130lb/60kg Capacity and 0.005lb/2g Readabil (GBK 130A) (YO-11119-85)
Adam GBK Industrial Scale, 130lb/60kg Capacity and 0.005lb/2g Readabil (GBK 130A) (YO-11119-86).
More Details
Adam Equipment Adam Gfk Industrial Scales
Buy Adam Equipment Adam Gfk Industrial Scales Items Including:
Adam GFK Industrial Scale, 1320lb/600kg Capacity And 0.1lb/50g Readabi
Adam GFK Industrial Scale, 165b/75kg Capacity And 0.01lb/5g Readabilit
Adam GFK Industrial Scale, 165b/75kg Capacity And 0.002lb/1g Readabili.
More Details
Adam Equipment Adam Highland Portable Balances
Buy Adam Equipment Adam Highland Portable Balances Items Including:
Adam Highland Portable Balance, 120g X 0.001g
Adam Highland Portable Balance, 300g X 0.01g
Adam Highland Portable Balance, 600g X .01g
Adam Highland Portable Balance, 120g X 0.001g (220 VAC)
Adam Highland Portable Balance, 150g X 0.005g (220 VAC).
More Details
Adam Equipment Adam Nimbus Analytical Balances
Buy Adam Equipment Adam Nimbus Analytical Balances Items Including:
Adam PGD 84e Analytical Balance, 80g with 0.1mg readability, graphic L (YO-11120-98)
Adam PGD 84e Analytical Balance, 80g with 0.1mg readability, graphic L (YO-11120-99).
More Details
Adam Equipment Adam Nimbus Toploading Balances
Buy Adam Equipment Adam Nimbus Toploading Balances Items Including:
Adam PGD 1003e Precision Toploader Balance, 1000g with 1mg readability (YO-11121-18)
Adam PGD 1003e Precision Toploader Balance, 1000g with 1mg readability (YO-11121-19).
More Details
Adam Equipment Afk Industrial Scales
Buy Adam Equipment Afk Industrial Scales Items Including:
Adam AFK Industrial Scale, 165lb/75kg Capacity and 0.01lb/5g Readabili (AFK 165A) (YO-11120-04)
Adam AFK Industrial Scale, 165lb/75kg Capacity and 0.01lb/5g Readabili (AFK 165A) (YO-11120-06).
More Details
Adam Equipment Balance and Scale Accessories
Buy Adam Equipment Balance and Scale Accessories Items Including:
Density Determination Kit for Adam Balances
Precision Toploading Balance Dust Cover for Adam Balances.
More Details
Adam Equipment Balance Anti-Theft Devices
Buy Adam Equipment Balance Anti-Theft Devices Items Including:
Kensington™-type security lock and cable (8030).
More Details
70618
results
<<<Previous
22
23
24
25
26
27
28
29
30
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Adam Afk Industrial Scales to Adam Equipment Balance Anti-Theft Devices
>
31125329-s-toc_2 31125329stoc2