Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 27
Adam Equipment Balance Cases
to
Adam Equipment Gfk Industrial Bench Scales
Image
Product Detail
More Detail
Adam Equipment Balance Cases
Buy Adam Equipment Balance Cases Items Including:
Hard Carrying Case with Lock and Strap for Adam Equipment balances (7959)
Carrying Case for Adam Equipment CPW Plus Industrial Scale
Hard Carrying Case with Lock and Strap for Adam CPWplus Balance (7954)
Hard Carrying Case w/Lock - CBK, CBC.
More Details
Adam Equipment Balance Density Kits
Buy Adam Equipment Balance Density Kits Items Including:
Density Determination Kit for Adam Equipment balances (7543)
Density Determination Kit for Adam Balances.
More Details
Adam Equipment Balance Dust Covers
Buy Adam Equipment Balance Dust Covers Items Including:
Analytical Balance Dust Cover for Adam Equipment balances (8003).
More Details
Adam Equipment Balance in-Use Covers
Buy Adam Equipment Balance in-Use Covers Items Including:
Balance In-use wet cover, Large for Adam Equipment balances (8031)
In use wet cover for CBD, CBC, CBW, CBK Qty
10
In use wet cover for CBD, CBC, CBW, CBK Qty
20
More Details
Adam Equipment Balance Interface Cables
Buy Adam Equipment Balance Interface Cables Items Including:
RS232 cable (YO-11120-93)
USB cable (YO-11120-94).
More Details
Adam Equipment Balance Printer Supplies
Buy Adam Equipment Balance Printer Supplies Items Including:
Printer paper for Adam Equipment printers, pack of 5 rolls (PRINTER PAPER)
Thermal Paper For Adam ATP (10 pack).
More Details
Adam Equipment Balance Thermal Printers
Buy Adam Equipment Balance Thermal Printers Items Including:
Thermal Printer for Balances with an RS-232 Interface (8023)
ATP Adam Thermal Printer
Replacement Paper for Thermal Printer 10000-38 (pack of 5) (1250).
More Details
Adam Equipment Balances
Buy Adam Equipment Balances Items Including:
Balance In-Use Wet Cover for Adam Equipment balances (7997)
Carrying Case for Adam Equipment CPW Plus Industrial Scale
Hard Carrying Case with Lock and Strap for Adam CPWplus Balance (7954)
Precision Toploading Balance Dust Cover for Adam Balances.
More Details
Adam Equipment Balances & Scales
Buy Adam Equipment Balances & Scales Items Including:
Balance In-Use Wet Cover for Adam Equipment balances (7997)
Analytical Balance Dust Cover for Adam Equipment balances (8003)
Carrying Case for Adam Equipment CPW Plus Industrial Scale
Adam GFK Industrial Scale, 165lb/75kg And 0.01lb/5g Readability.
More Details
Adam Equipment Balances Accessories
Buy Adam Equipment Balances Accessories Items Including:
Density Determination Kit for Adam Equipment balances (7543)
Density Determination Kit for Adam Balances.
More Details
Adam Equipment Balances Analytical
Buy Adam Equipment Balances Analytical Items Including:
Analytical Balance Dust Cover for Adam Equipment balances (8003)
Adam PGD 84e Analytical Balance, 80g with 0.1mg readability, graphic L (YO-11120-98)
Adam PGD 84e Analytical Balance, 80g with 0.1mg readability, graphic L (YO-11120-99).
More Details
Adam Equipment Balances Industrial and Scales
Buy Adam Equipment Balances Industrial and Scales Items Including:
Carrying Case for Adam Equipment CPW Plus Industrial Scale
Adam CPW Plus Industrial Scale, 35 kg x 0.01 kg, 115 VAC
Adam CPW Plus Industrial Scale, 150 kg x 0.05 kg, 115 VAC
Adam CPW Plus Industrial Scale, 150 kg x 0.05 kg, 230 VAC.
More Details
Adam Equipment Cbc Counting B
Buy Adam Equipment Cbc Counting B Items Including:
Adam CBC Counting Scale, 4 Kg X 0.1g
Adam CBC Counting Scale, 4 Kg X 0.1g, 220 VAC
Adam CBC Counting Scale, 8 Kg X 0.2g, 220 VAC
Adam CBC Counting Scale, 32 Kg X 1g
Adam CBC Counting Scale, 32 Kg X 1g, 220 VAC
More Details
Adam Equipment Cbc Counting Scales
Buy Adam Equipment Cbc Counting Scales Items Including:
Adam CBC Counting Scale, 4 Kg X 0.1g
Adam CBC Counting Scale, 4 Kg X 0.1g, 220 VAC
Adam CBC Counting Scale, 8 Kg X 0.2g, 220 VAC
Adam CBC Counting Scale, 32 Kg X 1g
Adam CBC Counting Scale, 32 Kg X 1g, 220 VAC
More Details
Adam Equipment Cbd Series Counting Scales
Buy Adam Equipment Cbd Series Counting Scales Items Including:
Adam CBD, Dual Platform Capable, 16lb/8000g x 0.0005lb/0.2g ,115V
Adam CBD, Dual Platform Capable, 16lb/8000g x 0.0005lb/0.2g ,220V
Adam CBD, Dual Platform Capable, USB, 8lb/4000g x 0.0002lb/0.1g ,115V.
More Details
Adam Equipment Core Portable Balances
Buy Adam Equipment Core Portable Balances Items Including:
Adam Core™ Portable Balance, 200g X 0.01g
Adam Core™ Portable Balance, 250g X 0.1g
Adam Core™ Portable Balance, 600g X 0.1g
Adam Core™ Portable Balance, 1500g X 0.1g
Adam Core™ Portable Balance, 2000g X 1g
More Details
Adam Equipment Counting Scales
Buy Adam Equipment Counting Scales Items Including:
Adam CBC Counting Scale, 4 Kg X 0.1g
Adam CBC Counting Scale, 4 Kg X 0.1g, 220 VAC
Adam CBC Counting Scale, 8 Kg X 0.2g, 220 VAC
Adam CBC Counting Scale, 32 Kg X 1g
Adam CBC Counting Scale, 32 Kg X 1g, 220 VAC
More Details
Adam Equipment Cpw Plus Industrial Scales
Buy Adam Equipment Cpw Plus Industrial Scales Items Including:
Carrying Case for Adam Equipment CPW Plus Industrial Scale
Adam CPW Plus Industrial Scale, 35 kg x 0.01 kg, 115 VAC
Adam CPW Plus Industrial Scale, 150 kg x 0.05 kg, 115 VAC
Adam CPW Plus Industrial Scale, 150 kg x 0.05 kg, 230 VAC.
More Details
Adam Equipment Cpwplus Industrial Scales
Buy Adam Equipment Cpwplus Industrial Scales Items Including:
Adam CPW Plus Industrial Scale, 35 kg x 0.01 kg, 230 VAC
Adam CPW Plus Industrial Scale, 75 kg x 0.02 kg, 115 VAC
Adam CPW Plus Industrial Scale, 150 kg x 0.05 kg, 115 VAC
Adam CPW Plus Industrial Scale, 150 kg x 0.05 kg, 230 VAC.
More Details
Adam Equipment Cqt Grain Scales
Buy Adam Equipment Cqt Grain Scales Items Including:
Adam CQT Grain Scale, 1750g Capacity and 0.05g Readability ,115V.
More Details
Adam Equipment Density Kit
Buy Adam Equipment Density Kit Items Including:
Density Determination Kit for Adam Equipment balances (7543)
Density Determination Kit for Adam Balances
Below Balance Hanger for Density Determination (7238).
More Details
Adam Equipment Digital Health Scales (Mdw-250l/300l)
Buy Adam Equipment Digital Health Scales (Mdw-250l/300l) Items Including:
Adam Digital Health Scale, MDW-250L, 550lb/250kg Capacity 115V
Adam Digital Health Scale, MDW-250L, 550lb/250kg Capacity 230V
Adam Digital Health Scale, MDW-300L, 600lb/300kg Capacity 115V.
More Details
Adam Equipment Dune Handheld Balance
Buy Adam Equipment Dune Handheld Balance Items Including:
DCT 201 Adam Dune™ Handheld Balance With 200g Capacity And 0.1g Readab
DCT 2000 Adam Dune™ Handheld Balance With 2000g Capacity And 1g Readab.
More Details
Adam Equipment Eclipse Toploading Balances
Buy Adam Equipment Eclipse Toploading Balances Items Including:
Adam PGD 253i Precision Toploader Balance, 250g w/1mg readability, gra (YO-11121-36)
Adam PGD 253i Precision Toploader Balance, 250g w/1mg readability, gra (YO-11121-37).
More Details
Adam Equipment Food and Portion Scales
Buy Adam Equipment Food and Portion Scales Items Including:
Adam WBW Portion Control Washdown Scale, 5A, With 5lb/2000g Capacity A
Adam WBW Portion Control Washdown Scale, 9A, With 9lb/4000g Capacity A
Adam WBW Portion Control Washdown Scale, 18A, With 18lb/8000g Capacity.
More Details
Adam Equipment Gbc Industrial Counting Scales
Buy Adam Equipment Gbc Industrial Counting Scales Items Including:
Adam GBC 35a Bench Adam GBD, 35lb/16kg Capacity, 0.001lb/0.5g Readabil (GBC 35a -115V)
Adam GBC 130a Bench Adam GBD, 130lb/60kg Capacity, 0.005lb/2g Readabil (GBC 130a -115V).
More Details
Adam Equipment Gbk Industrial Scales
Buy Adam Equipment Gbk Industrial Scales Items Including:
Adam GBK Industrial Scale, 130lb/60kg Capacity and 0.005lb/2g Readabil (GBK 130A) (YO-11119-85)
Adam GBK Industrial Scale, 130lb/60kg Capacity and 0.005lb/2g Readabil (GBK 130A) (YO-11119-86).
More Details
Adam Equipment Gfc Industrial Counting Scales
Buy Adam Equipment Gfc Industrial Counting Scales Items Including:
Adam GFC 165a Floor Adam GBD, 165lb/75kg Capacity, 0.01lb/5g Readabili (GFC 165a -115V)
Adam GFC 165a Floor Adam GBD, 165lb/75kg Capacity, 0.01lb/5g Readabili (GFC 165a -220V).
More Details
Adam Equipment Gfk Industrial
Buy Adam Equipment Gfk Industrial Items Including:
Adam GFK Industrial Scale, 1320lb/600kg Capacity And 0.1lb/50g Readabi
Adam GFK Industrial Scale, 165b/75kg Capacity And 0.01lb/5g Readabilit
Adam GFK Industrial Scale, 165b/75kg Capacity And 0.002lb/1g Readabili.
More Details
Adam Equipment Gfk Industrial Bench Scales
Buy Adam Equipment Gfk Industrial Bench Scales Items Including:
Adam GFK Industrial Scale, 1320lb/600kg Capacity And 0.1lb/50g Readabi
Adam GFK Industrial Scale, 165b/75kg Capacity And 0.01lb/5g Readabilit
Adam GFK Industrial Scale, 165b/75kg Capacity And 0.002lb/1g Readabili.
More Details
70618
results
<<<Previous
23
24
25
26
27
28
29
30
31
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
41
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Adam Equipment Balance Cases to Adam Equipment Gfk Industrial Bench Scales
>
31125329-s-toc_2 31125329stoc2