Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 29
Adam Pmb Moisture Determination Balances
to
Adapters Hose
Image
Product Detail
More Detail
Adam Pmb Moisture Determination Balances
Buy Adam Pmb Moisture Determination Balances Items Including:
Adam PMB Moisture Determination Balance, 50g X 1mg/0.01%, 110 VAC
Adam PMB Moisture Determination Balance, 50g X 1mg/0.01%, 220 VAC
Adam PMB Moisture Determination Balance, 200g X 01g/0.05%, 110 VAC.
More Details
Adam Portable Balances
Buy Adam Portable Balances Items Including:
Adam Core™ Portable Balance, 200g X 0.01g.
More Details
Adam Portable Toploading Balances
Buy Adam Portable Toploading Balances Items Including:
Adam Core™ Portable Balance, 200g X 0.01g
Adam Core™ Portable Balance, 250g X 0.1g
Adam Core™ Portable Balance, 600g X 0.1g
Adam Core™ Portable Balance, 1500g X 0.1g
Adam Core™ Portable Balance, 2000g X 1g
More Details
Adam Toploading Balances
Buy Adam Toploading Balances Items Including:
Adam PGW Precision Toploading Balance, 150 g x 0.001 g - 115 VAC
Adam PGD 1003e Precision Toploader Balance, 1000g with 1mg readability (YO-11121-18)
Adam PGD 1003e Precision Toploader Balance, 1000g with 1mg readability (YO-11121-19).
More Details
Adam Warrior Washdown Industrial Scales
Buy Adam Warrior Washdown Industrial Scales Items Including:
Adam Warrior Washdown Industrial Scale, WSK 16a, 16lb/8kg Capacity and
Adam Warrior Washdown Industrial Scale, 16a, 16lb/8kg Capacity and 0.0
Adam Warrior Washdown Industrial Scale, 165lb/75kg Capacity and 0.01lb (WFK 165A) (YO-11120-29).
More Details
Adam Wbw Portion Control Washdown Scales
Buy Adam Wbw Portion Control Washdown Scales Items Including:
Adam WBW Portion Control Washdown Scale, 5A, With 5lb/2000g Capacity A
Adam WBW Portion Control Washdown Scale, 9A, With 9lb/4000g Capacity A
Adam WBW Portion Control Washdown Scale, 18A, With 18lb/8000g Capacity.
More Details
Adampak High Temperature Labels
Buy Adampak High Temperature Labels Items Including:
Five-Point temperature indicating labels; temperature points 105-130d (YO-90303-20)
Five-Point temperature indicating labels; temperature points 140-180d (YO-90303-22).
More Details
Adams Equipment Scales
Buy Adams Equipment Scales Items Including:
Printer paper for Adam Equipment printers, pack of 5 rolls (PRINTER PAPER)
Adam PMB Moisture Determination Balance, 200g X 01g/0.05%, 110 VAC
Adam PMB Moisture Determination Balance, 200g X 01g/0.05%, 220 VAC.
More Details
Adams Glass Centrifuge Tubes
Buy Adams Glass Centrifuge Tubes Items Including:
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 5 ml, 125 (99502-5)
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 10 ml, 12 (99502-10)
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 15 ml, 12 (99502-15).
More Details
Adams Scales
Buy Adams Scales Items Including:
Adam LBK Compact Industrial Bench Scale, 30 kg, 115VAC
Adam LBK Compact Industrial Bench Scale, 3 kg, 230VAC
Adam LBK Compact Industrial Bench Scale, 12 kg, 230VAC
Adam CBC Counting Scale, 4 Kg X 0.1g
Adam CBC Counting Scale, 4 Kg X 0.1g, 220 VAC.
More Details
Adapter 110
Buy Adapter 110 Items Including:
Power Supply/Adapter (110-00-18T)
Power Supply/Adapter (110-00-08T)
Power adapter, 110 VAC
Power Adapter 110/240V for 1500 pH Meter (YO-99855-90)
Replacement AC adapter, 110 AC for 35710-35 (DT246 DC 6V)
More Details
Adapter 6000
Buy Adapter 6000 Items Including:
Replacement tube adapters, for 0.5-mL and 0.6 mL tubes (YO-17405-80)
Replacement tube adapters, for 0.25-mL and 0.4 mL tubes (YO-17420-67).
More Details
Adapter Bsp Npt
Buy Adapter Bsp Npt Items Including:
Threaded adapter, stainless steel, 1/8" female NPT to 1/8" male BSPT (YO-31830-40)
Threaded adapter, stainless steel, 1/4" female NPT to 1/4" male BSPT (YO-31830-41).
More Details
Adapter Camera
Buy Adapter Camera Items Including:
C-mount adapter for Canon 35-mm camera (MA150/50+MA512+T2-1)
C-mount adapter for Canon EOS 35-mm camera (MA150/50+MA512+T2-9)
C-mount adapter for 1/3" chip camera (MA151/35/03)
10:1 Stereozoom Microscope 35-mm Camera Adapter, Canon EOS (T2-9).
More Details
Adapter Canon Eos
Buy Adapter Canon Eos Items Including:
10:1 Stereozoom Microscope 35-mm Camera Adapter, Canon EOS (T2-9).
More Details
Adapter Fitting Flare
Buy Adapter Fitting Flare Items Including:
Flare compression pipe adapter, 1/4" x 1/8", PTFE, Each (YO-30490-18)
Flare compression pipe adapter, 1/4" x 1/4", PTFE, Each (YO-30490-20)
Flare compression male branch adapter, 1/4" x 1/8", PTFE, Each (YO-30490-88).
More Details
Adapter for Compression Nuts
Buy Adapter for Compression Nuts Items Including:
Drum Adapter System Kit, 330mm tube and 1/2" compression nut connectio (YO-31608-00)
Drum Adapter System Kit, 902mm tube and 1/2" compression nut connectio (YO-31608-01).
More Details
Adapter Heads
Buy Adapter Heads Items Including:
316 stainless steel adapter head; 3/4" NPT(F) (156094)
316 stainless steel adapter head; 1" NPT(F) (156095)
316 stainless steel adapter head; 3" NPT(F) (156158)
Masterflex I/P NEMA Type 56 C pump head adapter, 3.1:1 adapter gear ra.
More Details
Adapter Male
Buy Adapter Male Items Including:
Compression male pipe adapter, 1/8" x 1/8", Nylon, 10 Pack (YO-06382-36)
Compression male pipe adapter, 1/4" x 1/8", Nylon, 10 Pack (YO-06382-37)
Compression male pipe adapter, 1/4" x 1/4", Nylon, 10 Pack (YO-06382-38).
More Details
Adapter Outlet
Buy Adapter Outlet Items Including:
Inlet/Outlet fitting, PP1-1/2" NPT(M) x 1 1/2" ID (YO-06450-84)
Push-to-connect three-way divider, 1/2" inlet x 3/8" outlets, 10/pack (YO-34007-26)
Outlet Adapter O-ring, Uhp25/43/62 (341016).
More Details
Adapter Printer Cable
Buy Adapter Printer Cable Items Including:
OHAUS RS-232 CABLE AND ADAPTER FOR COMPACT THERMAL PRINTER 11901-47 (80252580)
Ohaus RS-232 Cable And Adapter For Compact Dot Matrix Printer 11901-59 (Accessory) (YO-11901-63)
OHAUS RS-232 ADAPTER FOR COMPACT DOT MATRIX PRINTER 11901-59 (Accessory).
More Details
Adapter Sleeves
Buy Adapter Sleeves Items Including:
Reducer sleeves, for 0.6 and 0.5-mL microcentrifuge tubes (YO-17701-54)
Reducer sleeves, for 0.4 and 0.25-mL microcentrifuge tubes (YO-17701-56).
More Details
Adapter Straight
Buy Adapter Straight Items Including:
Compression fittings, male pipe adapter, straight, Black PP, 1/4" OD x (YO-06444-01)
Compression fittings, male pipe adapter, straight, Black PP, 3/8" OD x (YO-06444-02).
More Details
Adapter Straight Pipe to Tube
Buy Adapter Straight Pipe to Tube Items Including:
Barbed fittings, NPT male pipe adapter, Natural PP, 1/8" NPT x 1/4" ID (YO-30610-36)
Barbed fittings, NPT male pipe adapter, Natural PP, 1/8" NPT x 3/8" ID (YO-30610-37).
More Details
Adapter Tube Female to Male
Buy Adapter Tube Female to Male Items Including:
Side arm injection port, 2 female, 1 male slip luer (YO-31507-83)
Luer fittings, 316 SS fittings, female luer x male slip (YO-31507-20)
316 SS fittings; female luer x male luer lock (YO-31507-21)
Luer Tee, Male-Male-Female, PP, 25/pk (YO-45500-75).
More Details
Adapters Adapter
Buy Adapters Adapter Items Including:
ETFE adapters, Adapter with 1/4-28 UNF Thread, 1/4 (YO-06471-92)
ETFE adapters, Adapter with 1/4-28 UNF Thread, 1/8 (YO-06471-93)
316 stainless steel adapter head; 3/4" NPT(F) (156094)
316 stainless steel adapter head; 1" NPT(F) (156095).
More Details
Adapters for Eppendorf Multi-Purpose Centrifuges
Buy Adapters for Eppendorf Multi-Purpose Centrifuges Items Including:
Adapter for fixed-angle and aerosol rotors; 0.25 to 0.4 mL, 6/pack
Adapter for fixed-angle and aerosol rotors; 0.2 PCR, 6/pack.
More Details
Adapters for Erp Modular Electrodes
Buy Adapters for Erp Modular Electrodes Items Including:
Submersion adapter, No ATC or solution ground (YO-27113-20)
In-line or submersion adapter with solution ground, No ATC (YO-27113-22)
In-line or submersion adapter, 100 Ohm ATC with solution ground (YO-27113-24).
More Details
Adapters for John Guest Polypropylene Fittings
Buy Adapters for John Guest Polypropylene Fittings Items Including:
Push-to-connect male adapter, polypropylene, 1/4" x 1/8" NPTM, 10/pack (YO-34007-42)
Push-to-connect male adapter, polypropylene, 1/4" x 1/4" NPTM, 10/pack (YO-34007-43).
More Details
Adapters Hose to Threaded Male Pipe
Buy Adapters Hose to Threaded Male Pipe Items Including:
Barbed fittings, NPT male pipe adapter, Natural PP, 1/8" NPT x 1/4" ID (YO-30610-36)
Barbed fittings, NPT male pipe adapter, Natural PP, 1/8" NPT x 3/8" ID (YO-30610-37).
More Details
70618
results
<<<Previous
25
26
27
28
29
30
31
32
33
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
41
42
43
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Adam Pmb Moisture Determination Balances to Adapters Hose to Threaded Male Pipe
>
31125329-s-toc_2 31125329stoc2