Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 3
A&D Stainless Steel Washdown Scales
to
a&D Weighing Fx-Iwp Waterproof Toploading Balances
Image
Product Detail
More Detail
A&D Stainless Steel Washdown Scales
Buy A&D Stainless Steel Washdown Scales Items Including:
A&D Portable Stainless Steel Washdown Scale, 1000 g (SK-1000WP)
A&D SW Washdown Industrial Scale, Stainless Steel Hi-Pressure Washdown (SW-15KS)
A&D SW Washdown Industrial Scale, Stainless Steel Hi-Pressure Washdown (SW-150KM).
More Details
A&D Sw Washdown Industrial Scales
Buy A&D Sw Washdown Industrial Scales Items Including:
A&D SW Washdown Industrial Scale, Stainless Steel Hi-Pressure Washdown (SW-15KS)
A&D SW Washdown Industrial Scale, Stainless Steel Hi-Pressure Washdown (SW-150KM).
More Details
A&D W Analytical Balances
Buy A&D W Analytical Balances Items Including:
Ohaus Pioneer Analytical Balance, 65 G X 0.1 Mg (115 VAC)
Ohaus Pioneer Analytical Balance, 110 G X 0.1 Mg (115 VAC)
Ohaus Pioneer Analytical Balance, 210 G X 0.1 Mg (115 VAC)
Ohaus Adventurer™ Pro Analytical Balance, 65 g x 0.0001 g, 115VAC, (in.
More Details
A&D W and Weighing Analytical Balances
Buy A&D W and Weighing Analytical Balances Items Including:
A&D Gemini Autocalibrating Analytical Balance, 120 G
A&D Orion Analytical Balance, 310 g x 0.1 mg, 120 VAC
A&D Orion Analytical Balance, 42/210 g x 0.01 /0.1 mg
A&D Galaxy HR-AZ Analytical Balance, 102g x 0.1mgwith Internal Calibra.
More Details
A&D W and Weighing Precision
Buy A&D W and Weighing Precision Items Including:
A&D AD-4212A Precision Weighing Sensor, 210g X 0.001g
A&D AD-4212A Precision Weighing Sensor, 1100g X 0.001g
A&D AD-4212A Precision Weighing Sensor, 110g X 0.1mg 230 VAC
A&D AD-4212A Precision Weighing Sensor, 210g X 0.001g 230 VAC.
More Details
A&D W Balance and Scale Accessories
Buy A&D W Balance and Scale Accessories Items Including:
24 Months Extended Warranty for All Mettler Toledo Balances (EW24)
Vistek™ Balance Isolation Platform, 8"W x 10"D
Disposable aluminum sample pans for Balances, pack of 250 (ALUMINUM PANS)
RS-232 interface for 11113-00 and -10 scales only.
More Details
A&D W Balances & Scales
Buy A&D W Balances & Scales Items Including:
Cole-Parmer Balance Draft Shield, Small 13"W x 13-1/2"H x 14-3/4"D (small)
Ohaus Defender 5000 Standard Industrial Scale Base, 60kgx0.01kg;large
Ohaus Defender 5000 Standard Industrial Scale Base, 150kgx0.02kg;xlarg.
More Details
A&D W General Accessories
Buy A&D W General Accessories Items Including:
General Purpose Probe 15FT/4.5M Long (TMX2G )
Horizontal pulse accessory; 110 VAC, 50/60 Hz
Label Kit Accessory
General-Purpose RTD Probe for Dual-Input RTD Thermometer with ANSI Con (850182)
801515 : ss General Purpose T/C- J Probe (YO-95001-82).
More Details
A&D W Moisture Balances
Buy A&D W Moisture Balances Items Including:
Ohaus MB Moisture Determination Balance, Mb23 110 G X 0.01 G 120VAC
Ohaus MB Moisture Determination Balance, Mb25 110 G X 0.005 G 120VAC
Mettler Toledo Deluxe Halogen Moisture Analyzer HR83, 81 g x 0.001 g (HR83)
Ohaus Aluminum Sample Pans, 80/Box.
More Details
A&D W Portable Balances
Buy A&D W Portable Balances Items Including:
Cole-Parmer Symmetry ED Portable Balance, 300g x 0.1g, 115VAC
Cole-Parmer Symmetry ED Portable Balance, 500g x 0.1g, 115VAC
Cole-Parmer Symmetry ED Portable Balance, 300g x 0.1g, 220VAC
Cole-Parmer Symmetry ED Portable Balance, 500g x 0.1g, 220VAC.
More Details
A&D W Toploading Balances
Buy A&D W Toploading Balances Items Including:
Cole-Parmer Symmetry PR-Precision Toploading Balance, 210 g x 0.001g - (YO-10000-11)
Cole-Parmer Symmetry PR-Precision Toploading Balance, 410 g x 0.001g - (YO-10000-12)
Cole-Parmer Symmetry PR-Precision Toploading Balance, 4200 g x 0.01g -.
More Details
A&D W Tuning Fork Vibration Viscometer
Buy A&D W Tuning Fork Vibration Viscometer Items Including:
Low range tuning fork vibration viscometer 120 VAC
High range tuning fork vibration viscometer 120 VAC
High range tuning fork vibration viscometer 240 VAC.
More Details
A&D W Viscometers
Buy A&D W Viscometers Items Including:
Viscometer Tube, Brass, for Saybolt Viscometer Bath (K22009)
Viscometer Tube, Stainless steel, for Saybolt Viscometer Bath (K22309)
Socket wrench for Saybolt Viscometer Bath (K22050)
Receiving flask, Pyrex glass, 60 mL, for Saybolt Viscometer Bath.
More Details
A&D Weighing
Buy A&D Weighing Items Including:
A&D AD-4212A Precision Weighing Sensor, 210g X 0.001g
A&D AD-4212A Precision Weighing Sensor, 1100g X 0.001g
A&D AD-4212A Precision Weighing Sensor, 110g X 0.1mg 230 VAC
A&D AD-4212A Precision Weighing Sensor, 210g X 0.001g 230 VAC.
More Details
A&D Weighing Ad-4212a Precision Weighing Sensor
Buy A&D Weighing Ad-4212a Precision Weighing Sensor Items Including:
A&D AD-4212A Precision Weighing Sensor, 210g X 0.001g
A&D AD-4212A Precision Weighing Sensor, 1100g X 0.001g
A&D AD-4212A Precision Weighing Sensor, 110g X 0.1mg 230 VAC
A&D AD-4212A Precision Weighing Sensor, 210g X 0.001g 230 VAC.
More Details
A&D Weighing Ad-4406 Weighing Indicator
Buy A&D Weighing Ad-4406 Weighing Indicator Items Including:
Ad-4406-03:Rs-422 with Relays for A&D Weighing Indicators.
More Details
A&D Weighing and Engineering Portable Balances
Buy A&D Weighing and Engineering Portable Balances Items Including:
A&D Portable Balance, 120 g x 0.01 g (SV-120)
A&D Portable Balance, 200 g, 0.01 g (SV-200)
Power Adapter, 220 VAC for A&D HC-i Portable Counting Scale
A&D Disposable Sample Pans for Mositure Analyzers 100pcs..
More Details
A&D Weighing and Weighing Analytical Balances
Buy A&D Weighing and Weighing Analytical Balances Items Including:
A&D ION Semi-Micro Balance, 250g x 0.01mg IntCal, Ionizer, Env.Sensors
A&D Gemini Autocalibrating Analytical Balance, 120 G
A&D Orion Analytical Balance, 310 g x 0.1 mg, 120 VAC
A&D Orion Analytical Balance, 42/210 g x 0.01 /0.1 mg.
More Details
A&D Weighing and Weighing Balance Accessories
Buy A&D Weighing and Weighing Balance Accessories Items Including:
Ad-4406-03:Rs-422 with Relays for A&D Weighing Indicators
In-Use Cover for A&D GF Series Balances
A&D Weighing DataLogger Stores Approx 5000 readings and uploads to Exc
Micro Aluminum Weighing Dishes, 6mm Dia
x 2.5mm H, .035ml, 250/pk.
More Details
A&D Weighing and Weighing Portable Balances
Buy A&D Weighing and Weighing Portable Balances Items Including:
A&D Eki Portable Balance, 120 g x 0.01 g
A&D Eki Portable Balance, 200 g x 0.01 g
A&D Eki Portable Balance, 1200 g capacity
A&D Eki Portable Balance, 2000 g capacity
A&D Eki Portable Balance, 3000 g capacity.
More Details
A&D Weighing and Weighing Scales
Buy A&D Weighing and Weighing Scales Items Including:
A&D AD-4212A Precision Weighing Sensor, 210g X 0.001g
A&D AD-4212A Precision Weighing Sensor, 1100g X 0.001g
A&D AD-4212A Precision Weighing Sensor, 110g X 0.1mg 230 VAC
A&D AD-4212A Precision Weighing Sensor, 210g X 0.001g 230 VAC.
More Details
A&D Weighing Balance Accessories
Buy A&D Weighing Balance Accessories Items Including:
In-Use Cover for A&D GF Series Balances
AND Moisture Determination Balance RS-232 Interface
Density Determination Kit for A&D Weighing HR-A/HR-AZ
Cover, SS weighing pan (Accessory)
USB Interface for A&D Weighing Newton Balance (EJ-02).
More Details
A&D Weighing Balance and Scale Accessories
Buy A&D Weighing Balance and Scale Accessories Items Including:
Statistical Printer for A&D Weighing Balances
Comparator relay output/buzzer for A&D Washdown Bench Scales (HV/W-03G)
AND Weighting RS232 with Comparator Relay Output for SW-Series Scales
AND Weighting RS422/485 Output for SW-Series Scales.
More Details
A&D Weighing Balances Industrial and Scales
Buy A&D Weighing Balances Industrial and Scales Items Including:
A&D HW Industrial Scale, 60 kg x 0.005 kg
A&D FG-K Industrial Scale, 60 lb x 0.01 lb with display column
A&D FG-K Industrial Scale, 150 lb x 0.02 lb with display column
A&D FG-K Industrial Scale, 300 lb x 0.1 lb with display column.
More Details
A&D Weighing Everest Eki Portable Balances
Buy A&D Weighing Everest Eki Portable Balances Items Including:
A&D Eki Portable Balance, 120 g x 0.01 g
A&D Eki Portable Balance, 1200 g capacity
A&D Eki Portable Balance, 2000 g capacity
A&D Eki Portable Balance, 3000 g capacity
A&D Eki Portable Balance, 12000 g capacity.
More Details
A&D Weighing Fg-K Industrial Scales
Buy A&D Weighing Fg-K Industrial Scales Items Including:
A&D FG-K Industrial Scale, 60 lb x 0.01 lb with display column
A&D FG-K Industrial Scale, 150 lb x 0.02 lb with display column.
More Details
A&D Weighing Food and Portion Scales
Buy A&D Weighing Food and Portion Scales Items Including:
A&D SJ Series Digital Portion Scale, 2000 G X 1 G
A&D SJ Series Digital Portion Scale, 12 Kg X 0.005 Kg
A&D Portable Stainless Steel Washdown Scale, 1000 g (SK-1000WP)
A&D Portable Stainless Steel Washdown Scale, 2000 g (SK-2000WP).
More Details
A&D Weighing Fsi Washdown Industrial Scales
Buy A&D Weighing Fsi Washdown Industrial Scales Items Including:
A&D FSi Washdown Industrial Scale, 6 kg (15 lb)
A&D FSi Washdown Industrial Scale, 15 kg (30 lb).
More Details
A&D Weighing Fx-I Toploading Balances
Buy A&D Weighing Fx-I Toploading Balances Items Including:
A&D FX-i Toploading Balance 122g X 0.001g Ext.Calibration, Comparator,.
More Details
A&D Weighing Fx-Iwp Waterproof Toploading Balances
Buy A&D Weighing Fx-Iwp Waterproof Toploading Balances Items Including:
A&D FX-iWP Toploading Balance, 1220g X 0.01g Ext.Calibration, Comparat (FX-1200IWP+230VAC)
A&D FX-iWP Toploading Balance, 2200g X 0.01g Ext.Calibration, Comparat (FX-2000IWP+230VAC).
More Details
70618
results
<<<Previous
1
2
3
4
5
6
7
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,354
A&D Stainless Steel Washdown Scales to a&D Weighing Fx-Iwp Waterproof Toploading Balances
>
31125329-s-toc_2 31125329stoc2