Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 300
Cole Compact
to
Cole Conductivity Calibration
Image
Product Detail
More Detail
Cole Compact
Buy Cole Compact Items Including:
Cole-Parmer Compact Sysring Pump 115 VAC
Cole-Parmer Compact Sysring Pump 230v
Cole-Parmer Computer Cable For 75900 Series Of Pumps 7 Foot Length
Cole-Parmer Compact Filtration System, PP/PVC
Cole-Parmer Ceramic Hot Plate, 4" x 4", 120V.
More Details
Cole Compact Color Meters
Buy Cole Compact Color Meters Items Including:
Compact Color Meter, 3-mm
Compact Color Meter, 5-mm, sapphire window
Compact Color Meter, 5-mm, sapphire window, software included
Compact Color Meter, 10-mm
Compact Color Meter, 10-mm, sapphire window
More Details
Cole Compact Digital Lab Mixer
Buy Cole Compact Digital Lab Mixer Items Including:
Compact square blade impeller; 1-1/4" dia
Compact Digital Mixer System, 50 to 2500 rpm, 115 VAC
Compact Digital Mixer System, 50 to 2500 rpm, 115/230 VAC with Plug Ad.
More Details
Cole Compact Electromagnetic Sieve Shaker
Buy Cole Compact Electromagnetic Sieve Shaker Items Including:
Compact Electromagnetic Sieve Shaker; 115 VAC, 60 Hz.
More Details
Cole Compact Gas Mass Flow Controllers
Buy Cole Compact Gas Mass Flow Controllers Items Including:
Thermal gas mass flow controller, to 10 sccm Air / N2, Alum (GFCS-010002)
Thermal gas mass flow controller, to 20 sccm Air / N2, Alum (GFCS-010003)
Thermal gas mass flow controller, to 50 sccm Air / N2, Alum (GFCS-010004).
More Details
Cole Compact Gas Mass Flowmeters
Buy Cole Compact Gas Mass Flowmeters Items Including:
Thermal Gas Mass Flowmeter, to 10 sccm Air / N2, Aluminum b (GFMS-010001)
Thermal Gas Mass Flowmeter, to 20 sccm Air / N2, Aluminum b (GFMS-010002)
Thermal Gas Mass Flowmeter, to 200 sccm Air / N2, 316SS bod (GFMS-010023).
More Details
Cole Compact Incubators
Buy Cole Compact Incubators Items Including:
Compact Incubator; 0.8 cu ft; 115V (YO-03608-16).
More Details
Cole Compact Low-Flow Switches
Buy Cole Compact Low-Flow Switches Items Including:
Compact low-flow switch for gases; SPST normally open, 25 cfh/60 cfh (YO-32778-30)
Compact low-flow switch for liquids; SPST normally open, 0.05 GPM (YO-32778-31)
Compact low-flow switch for gases; SPST normally open, 2.5 cfm/5.6 cfm (YO-32778-34).
More Details
Cole Compact Outlet Strips
Buy Cole Compact Outlet Strips Items Including:
Compact 10-outlet strip, 15-ft cord (UL210BD).
More Details
Cole Compact Pressure Transducer
Buy Cole Compact Pressure Transducer Items Including:
CP Pressure Transmitter, 0 To 50 PSIG; 1 To 5 VDC Output, 1/4" NPT(M).
More Details
Cole Compact Stirrers
Buy Cole Compact Stirrers Items Including:
Air- or Water-Driven Magnetic Stirrer (204015)
Ika Ultra-Flat Lab Disc Stirrer Sunny Side Up (3362300)
Ika Ultra-Flat Lab Disc Stirrer Orange (3578000)
Ika Ultra-Flat Lab Disc Stirrer Green (3578200)
Ika Ultra-Flat Lab Disc Stirrer Pink (3578400).
More Details
Cole Compact Ultrasonic Cleaners
Buy Cole Compact Ultrasonic Cleaners Items Including:
Cole-Parmer Low-Cost Ultrasonic Cleaner with Timer, 117 VAC (YO-08848-10)
Cole-Parmer Low-Cost Ultrasonic Cleaner with Timer, 220 VAC (YO-08848-15)
Compact Ultrasonic Test Sieve Cleaner, 115 VAC
Plastic lid for 59987-35 and -36 Ultrasonic Cleaners.
More Details
Cole Compact Usb Dataloggers
Buy Cole Compact Usb Dataloggers Items Including:
USB Temperature/Humidity Data Logger, 16,000+ readings, 1 year battery
USB Voltage Data Logger, Process, 32,000+ readings, 1 year battery lif
Temperature & Humidity and Dew Point Data Logger with LCD Display, 16,.
More Details
Cole Compact Variable-S
Buy Cole Compact Variable-S Items Including:
Compact Variable-Speed Universal Centrifuge Tube Adapter for 15-mL Fix (YO-17306-60)
Compact Variable-Speed Universal Centrifuge Tube Adapter for 50-mL Fix (YO-17306-64)
Standard Duty Dry Vacuum Diaphragm Pump, 43 L/min (1.6 CFM), 24 VDC.
More Details
Cole Compact Variable-Speed Universal Centrifuge
Buy Cole Compact Variable-Speed Universal Centrifuge Items Including:
Compact Variable-Speed Universal Centrifuge Tube Adapter for 15-mL Fix (YO-17306-60)
Compact Variable-Speed Universal Centrifuge Tube Adapter for 50-mL Fix (YO-17306-64)
Fixed-angle Rotor, 18 x 1.5-mL tubes (YO-17306-56).
More Details
Cole Compact/Oem Vacuum Pumps
Buy Cole Compact/Oem Vacuum Pumps Items Including:
Compact/oem Vacuum Pump, 12 VDC, 1.4 LPM, 8.8
Compact/OEM bellows vacuum/pressure pump, 0.4 LPM 5.9" Hg/6.8 psi, 3/6
Compact/OEM bellows pressure pump, 0.5 LPM 5.8 psi, 3 VDC
Dry rotary vane vacuum pump, 4.0 cfm, 220 VAC (0523-V550AQ-G588DX).
More Details
Cole Complete Drum Pump Systems
Buy Cole Complete Drum Pump Systems Items Including:
Pump, Motorized Drum Pump Package, Designed for Water Treatement Chemi (9430-110V) (YO-70614-00)
Pump, Motorized Drum Pump Package, Designed for Water Treatement Chemi (9431-230V) (YO-70614-01).
More Details
Cole Complete Turbomolecular System
Buy Cole Complete Turbomolecular System Items Including:
Complete turbomolecular system, 30 L/S, 115/208 VAC, 50/60 Hz.
More Details
Cole Compound Microscopes
Buy Cole Compound Microscopes Items Including:
Cole-Parmer professional compound microscope; monocular, achromatic ob (YO-48923-00)
Cole-Parmer professional compound microscope; monocular, achromatic ob (YO-48923-05)
Compound Microscope with Infinity-Corrected Optics, Binocular, 110 to.
More Details
Cole Compound Vials
Buy Cole Compound Vials Items Including:
Compound vials, 14.75 x 19 mm, 0.95 mL, clear glass, 144/case (YO-03757-14)
Compound vials, 14.75 x 22 mm, 1.25 mL, clear glass, 144/case (YO-03757-16)
Compound vials, 14.75 x 26 mm, 1.85 mL, clear glass, 144/case (YO-03757-18).
More Details
Cole Compressed
Buy Cole Compressed Items Including:
Compression nut assemblies (include ferrules), 1/8", PP, 10 per pack (YO-06380-05)
Compression nut assembly (include ferrules), PVDF, 1/8" OD (YO-06380-60)
Straight compression union, polypropylene, 1/4" OD, 10 per pack (YO-06381-10).
More Details
Cole Compression Fittings
Buy Cole Compression Fittings Items Including:
Inserts for soft tubing for compression fittings, white PP, 5/16" OD x (YO-06391-20)
Compression fitting; probe diameter, 3/16"; 316 SS; 1/8"NPT (M) (3-2 FH4BZ-SS-C).
More Details
Cole Compression Tee
Buy Cole Compression Tee Items Including:
Union Tees (YO-06382-05)
Union Tees (YO-06382-10)
Union Tees (YO-06382-20)
Union Tee, 1/8", Nylon, 10 Pack (YO-06382-31)
Union Tee, 1/4", Nylon, 10 Pack (YO-06382-32)
More Details
Cole Compression Union Elbow
Buy Cole Compression Union Elbow Items Including:
Compression Union Elbow, 3/8", nylon, 10 per pack (YO-06382-22)
Compression Fitting, Union Elbow, 316 SS, 1/4" OD (YO-31406-07)
Union Elbow, Stainless Steel, 5/8", Each (YO-31406-57)
Union Elbow, Brass, 5/8", Each (YO-31412-60).
More Details
Cole Compression-to-Compression Fittings
Buy Cole Compression-to-Compression Fittings Items Including:
Compression nut assemblies (include ferrules), 1/8", PP, 10 per pack (YO-06380-05)
Compression nut assembly (include ferrules), PVDF, 1/8" OD (YO-06380-60)
Straight compression union, polypropylene, 1/4" OD, 10 per pack (YO-06381-10).
More Details
Cole Compression-to-Threaded Adapters
Buy Cole Compression-to-Threaded Adapters Items Including:
Compression Elbow Threaded Adapter, Stainless Steel, 1/8" x 1/8", Each (YO-31406-75)
Compression Elbow Threaded Adapter, Stainless Steel, 1/8" x 1/4", Each (YO-31406-76).
More Details
Cole Computer
Buy Cole Computer Items Including:
Cole-Parmer Computer Cable For 75900 Series Of Pumps 7 Foot Length
Cole-Parmer Computer Cable For 75900 Series Of Pumps 25 Foot Length
RS-232 interface cable; connects L/S computer-compatible drives to PC, (YO-07550-64).
More Details
Cole Con
Buy Cole Con Items Including:
pH Electrode, Fermentation, 12 x 225 mm, Wick Junction, ATC, 4-pin con (YO-29045-04)
pH Electrode, Fermentation, 12 x 225 mm, Open Junction, ATC, 4-pin con (YO-29045-14).
More Details
Cole Conductive Shoe Covers
Buy Cole Conductive Shoe Covers Items Including:
Conductive Shoe Cover, Polypropylene, Skid-free.
More Details
Cole Conductivity Calibration
Buy Cole Conductivity Calibration Items Including:
Cole-Parmer conductivity standard, 1000 µS, 16 ounces
Cole-Parmer conductivity standard, 100,000 µS, 16 ounces
Cole-Parmer conductivity standard, 1 µS, 16 ounces, individually te
Cole-Parmer conductivity standard, 5 µS, 16 ounces, individually te.
More Details
70618
results
<<<Previous
296
297
298
299
300
301
302
303
304
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
100
200
210
220
230
240
250
260
270
280
285
286
287
288
289
290
291
292
293
294
295
296
297
298
299
300
301
302
303
304
305
306
307
308
309
310
311
312
313
314
320
330
340
350
360
370
380
390
400
500
600
700
800
900
1,000
1,100
1,200
2,000
2,354
Cole Compact to Cole Conductivity Calibration
>
31125329-s-toc_2 31125329stoc2