Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 40
Advantec Extraction Thimble
to
Advantec Gridded Surface 37 Mm Monitorss
Image
Product Detail
More Detail
Advantec Extraction Thimbles, Cellulose
Buy Advantec Extraction Thimbles, Cellulose Items Including:
Advantec Extraction Thimbles, Cellulose, 43 mm OD x 123 mm H; 25/pack
Advantec Extraction Thimbles, Cellulose, 30 mm OD x 100 mm H; 25/pack
Advantec Extraction Thimbles, Cellulose, 25 mm OD x 100 mm H; 25/pack.
More Details
Advantec Extraction Thimbles, Glass Fiber
Buy Advantec Extraction Thimbles, Glass Fiber Items Including:
Advantec Extraction Thimbles, Glass Fiber, 19 mm OD x 90 mm H; 25/pack
Advantec Extraction Thimbles, Glass Fiber, 30 mm OD x 80 mm H; 25/pack (N086R30X80MM)
Advantec Extraction Thimbles, Glass Fiber, 30 mm OD x 80 mm H; 25/pack (N088R30X80MM).
More Details
Advantec Extraction Thimbles, Quartz Fiber
Buy Advantec Extraction Thimbles, Quartz Fiber Items Including:
Advantec Extraction Thimbles, Quartz Fiber, 19 mm OD x 90 mm H; 25/pac
Advantec Extraction Thimbles, Quartz Fiber, 25 mm OD x 100 mm H; 25/pa
Advantec Extraction Thimbles, Quartz Fiber, 30 mm OD x 100 mm H; 25/pa.
More Details
Advantec Filter
Buy Advantec Filter Items Including:
Advantec Grade No
1 Qualitative Filter Paper; 5.5 cm dia.; pack of 10
Advantec Grade No
1 Qualitative Filter Paper; 7.0 cm dia.; pack of 10
Advantec Grade No
More Details
Advantec Filter Membranes
Buy Advantec Filter Membranes Items Including:
Advantec Sterile Mixed Cellulose Ester Filter Membranes; 47mm dia; 0.6 (YO-06644-06)
Advantec Sterile Mixed Cellulose Ester Filter Membranes; 47mm dia; 0.4 (YO-06644-08).
More Details
Advantec Filter Papers
Buy Advantec Filter Papers Items Including:
Advantec Grade No
1 Qualitative Filter Paper; 5.5 cm dia.; pack of 10
Advantec Grade No
1 Qualitative Filter Paper; 7.0 cm dia.; pack of 10
Advantec Grade No
More Details
Advantec Filter Sterilizing
Buy Advantec Filter Sterilizing Items Including:
Advantec Glass Sterility Test Filter Holder for 47-mm Membranes, Sinte
Advantec Sterile Mixed Cellulose Ester Filter Membranes; 47mm dia; 0.6 (YO-06644-06)
Advantec Sterile Mixed Cellulose Ester Filter Membranes; 47mm dia; 0.4 (YO-06644-08).
More Details
Advantec Filtration
Buy Advantec Filtration Items Including:
Advantec Grade No
1 Qualitative Filter Paper; 5.5 cm dia.; pack of 10
Advantec Grade No
1 Qualitative Filter Paper; 7.0 cm dia.; pack of 10
Advantec Grade No
More Details
Advantec Fraction Collectors
Buy Advantec Fraction Collectors Items Including:
Advantec Microcomputer Controlled Fraction Collector, 110-240 VAC.
More Details
Advantec Glas Fiber Filters
Buy Advantec Glas Fiber Filters Items Including:
Advantec Grade GA55 Glass Fiber Filters, 125mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 47mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 90mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 125mm dia; 100/box.
More Details
Advantec Glass Air Sampling Filters
Buy Advantec Glass Air Sampling Filters Items Including:
Advantec Air Sampling Filters, Glass Fiber, 75-mm dia disc; pack of 10
Advantec Air Sampling Filters, Composite, 25-mm dia disc; pack of 100.
Whatman Air Sampling Filters, EPM, 4.7 cm Dia
Advantec Grade GA55 Glass Fiber Filters, 47mm dia; 100/box.
More Details
Advantec Glass and Quartz Fiber Filters
Buy Advantec Glass and Quartz Fiber Filters Items Including:
Quartz fiber filter, QR200, 55-mm dia; box of 50.
Glass fiber filter, GA55, 42.5-mm dia; box of 100
Glass fiber filter, GA55, 110-mm dia; box of 100
Glass fiber filter, GD120, 110-mm dia; box of 50
Glass fiber filter, GF75, 110-mm dia; box of 100.
More Details
Advantec Glass Fiber Filter Sheets
Buy Advantec Glass Fiber Filter Sheets Items Including:
Advantec Grade GA55 Glass Fiber Filters, 125mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 47mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 90mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 125mm dia; 100/box.
More Details
Advantec Glass Fiber Filters
Buy Advantec Glass Fiber Filters Items Including:
Advantec Grade GA55 Glass Fiber Filters, 125mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 47mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 90mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 125mm dia; 100/box.
More Details
Advantec Glass Fiber Thimbles
Buy Advantec Glass Fiber Thimbles Items Including:
Advantec Extraction Thimbles, Glass Fiber, 19 mm OD x 90 mm H; 25/pack
Advantec Extraction Thimbles, Glass Fiber, 30 mm OD x 80 mm H; 25/pack (N086R30X80MM)
Advantec Extraction Thimbles, Glass Fiber, 30 mm OD x 80 mm H; 25/pack (N088R30X80MM).
More Details
Advantec Grade Dp70 Glass Fiber Filters
Buy Advantec Grade Dp70 Glass Fiber Filters Items Including:
Advantec Grade DP70 Glass Fiber Filters; 47mm dia; 50/box
Advantec Grade DP70 Glass Fiber Filters; 125mm dia; 50/box
Advantec Grade DP70 Glass Fiber Filters; 142mm dia; 50/box
Advantec Grade DP70 Glass Fiber Filters; 293mm dia; 50/box.
More Details
Advantec Grade Ga100 Glass Fiber Filters
Buy Advantec Grade Ga100 Glass Fiber Filters Items Including:
Advantec Grade GA100 Glass Fiber Filters; 47mm dia; 100/box
Advantec Grade GA100 Glass Fiber Filters; 25mm dia; 100/box
Advantec Grade GA100 Glass Fiber Filters; 125mm dia; 100/box
Advantec Grade GA100 Glass Fiber Filters; 142mm dia; 100/box.
More Details
Advantec Grade Ga200 Glass Fiber Filters
Buy Advantec Grade Ga200 Glass Fiber Filters Items Including:
Advantec Grade GA200 Glass Fiber Filters; 25mm dia; 50/box
Advantec Grade GA200 Glass Fiber Filters; 125mm dia; 50/box
Advantec Grade GA200 Glass Fiber Filters; 142mm dia; 50/box
Advantec Grade GA200 Glass Fiber Filters; 293mm dia; 50/box.
More Details
Advantec Grade Ga55 Glass Fiber Filters
Buy Advantec Grade Ga55 Glass Fiber Filters Items Including:
Advantec Grade GA55 Glass Fiber Filters, 47mm dia; 100/box
Advantec Grade GA55 Glass Fiber Filters; 100/box
Advantec Grade GA55 Glass Fiber Filters, 125mm dia; 100/box
Advantec Grade GA55 Glass Fiber Filters, 25mm dia; 100/box.
More Details
Advantec Grade Gb100r Glass Fiber Filters
Buy Advantec Grade Gb100r Glass Fiber Filters Items Including:
Advantec Grade GB100R Glass Fiber Filters; 90mm dia; 100/box
Advantec Grade GB100R Glass Fiber Filters; 25mm dia; 100/box
Advantec Grade GB100R Glass Fiber Filters; 125mm dia; 100/box
Advantec Grade GB100R Glass Fiber Filters; 142mm dia; 100/box.
More Details
Advantec Grade Gb140 Glass Fiber Filters
Buy Advantec Grade Gb140 Glass Fiber Filters Items Including:
Advantec Grade GB140 Glass Fiber Filters; 47mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 90mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 125mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 142mm dia; 100/box.
More Details
Advantec Grade Gc50 Glass Fiber Filters
Buy Advantec Grade Gc50 Glass Fiber Filters Items Including:
Advantec Grade GC50 Glass Fiber Filters; 47mm dia; 100/box
Advantec Grade GC50 Glass Fiber Filters; 90mm dia; 100/box
Advantec Grade GC50 Glass Fiber Filters; 125mm dia; 100/box
Advantec Grade GC50 Glass Fiber Filters; 142mm dia; 100/box.
More Details
Advantec Grade Gc90 Glass Fiber Filters
Buy Advantec Grade Gc90 Glass Fiber Filters Items Including:
Advantec Grade GC90 Glass Fiber Filters; 293mm dia; 50/box
Advantec Grade GC90 Glass Fiber Filters; 90mm dia; 100/box
Advantec Grade GC90 Glass Fiber Filters; 25mm dia; 100/box
Advantec Grade GC90 Glass Fiber Filters; 125mm dia; 100/box.
More Details
Advantec Grade Gd120 Glass Fiber Filters
Buy Advantec Grade Gd120 Glass Fiber Filters Items Including:
Advantec Grade GD120 Glass Fiber Filters; 47mm dia; 50/box
Advantec Grade GD120 Glass Fiber Filters; 90mm dia; 50/box
Advantec Grade GD120 Glass Fiber Filters; 125mm dia; 50/box
Advantec Grade GD120 Glass Fiber Filters; 293mm dia; 50/box.
More Details
Advantec Grade Gf75 Glass Fiber Filters
Buy Advantec Grade Gf75 Glass Fiber Filters Items Including:
Advantec Grade GF75 Glass Fiber Filters; 47mm dia; 100/box
Advantec Grade GF75 Glass Fiber Filters; 90mm dia; 100/box
Advantec Grade GF75 Glass Fiber Filters; 125mm dia; 100/box
Advantec Grade GF75 Glass Fiber Filters; 142mm dia; 100/box.
More Details
Advantec Grade Gs25 Glass Fiber Filters
Buy Advantec Grade Gs25 Glass Fiber Filters Items Including:
Advantec Grade GS25 Glass Fiber Filters; 293mm dia; 50/box
Advantec Grade GS25 Glass Fiber Filters; 25mm dia; 100/box
Advantec Grade GS25 Glass Fiber Filters; 47mm dia; 100/box
Advantec Grade GS25 Glass Fiber Filters; 125mm dia; 100/box.
More Details
Advantec Grade Qr100 Quartz Fiber Filters
Buy Advantec Grade Qr100 Quartz Fiber Filters Items Including:
Advantec Grade QR100 Quartz Fiber Filters, 25-mm dia; box of 100.
Advantec Grade QR100 Quartz Fiber Filters, QR100, 90-mm dia; box of 10
Advantec Grade QR100 Quartz Fiber Filters, 125-mm dia; box of 100..
More Details
Advantec Grade Qr200 Quartz Fiber Filters
Buy Advantec Grade Qr200 Quartz Fiber Filters Items Including:
Advantec Grade QR200 Quartz Fiber Filters, 25-mm dia; box of 100.
Advantec Grade QR200 Quartz Fiber Filters, 47-mm dia; box of 50.
Advantec Grade QR200 Quartz Fiber Filters, 90-mm dia; box of 50..
More Details
Advantec Gridded Surface 37 Mm Monitors
Buy Advantec Gridded Surface 37 Mm Monitors Items Including:
Sterile two-piece monitor, gridded surface, 0.45 um, 50/pack (YO-02935-20)
Mixed Cellulose Ester (MCE) membrane for asbestos monitoring; 37-mm di (YO-06639-72)
Mixed Cellulose Ester (MCE) membrane for asbestos monitoring; 37-mm di (YO-06639-74).
More Details
Advantec Gridded Surface 37 Mm Monitorss
Buy Advantec Gridded Surface 37 Mm Monitorss Items Including:
Three-piece monitor, gridded surface, 0.80 um, 50/pack (YO-02935-10)
Three-piece monitor, gridded surface, 0.80 um, 50/pack (YO-02935-12)
Sterile two-piece monitor, gridded surface, 0.45 um, 50/pack (YO-02935-20).
More Details
70618
results
<<<Previous
36
37
38
39
40
41
42
43
44
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
20
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
53
54
60
70
80
90
100
110
120
130
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Advantec Extraction Thimble to Advantec Gridded Surface 37 Mm Monitorss
>
31125329-s-toc_2 31125329stoc2